1x1 image pixel for data collection
1064Defender Raman Analyzer
1064Defender Raman Analyzer
1064Defender Raman Analyzer
1064Defender Raman Analyzer
1064Defender Raman Analyzer
1064Defender Raman Analyzer
1064Defender Raman Analyzer
Thermo Scientific™
1064Defender Raman Analyzer
The Thermo Scientific 1064Defender Raman Analyzer removes the uncertainty from chemical and narcotics identification and delivers definitive answers with maximum flexibility.
Have Questions?
Catalog number 1064DEFENDER
Price (USD)
-
Request A Quote
Perform narcotics identification and chemical identification safely and quickly with the Thermo Scientific 1064Defender Raman Analyzer. The handheld 1064Defender analyzer is a truly flexible tool to meet your chemical identification needs. By offering user configurable scan profiles and expandable libraries, the 1064Defender Raman Analyzer removes the guess work and provides actionable results for confident and efficient decision making.
Ensure user safety with touchless, point and shoot scanning
  • Enhanced identification of fluorescing materials safeguards against the dangerous exposure
  • Screener mode takes the guess work out of identifying substances by delivering results as either an alarm, warning, or clear to ensuring users know what actions to take
 User flexibility—through a customizable interface—ensures you and the instrument are always in sync.
  • 3 levels of control – administrator, operator and manager
  • 2 different scan modes for advanced users – ID and Screener modes
  • Modular and flexible - the device is future-proofed against the changing threat environment for users and the communites they serve
 Solid chain of custody
  • GPS and Digital Camera for time and location stamping
  • Unique user ID and password, ensure your data is locked and safe
  • User generated meta data
  • Instrument logs who scanned what substances and when
A comprehensive and expandable library of controlled substances, cutting agents, and precursors
  • User defined libraries
  • Regular updates to ensure you stay current with emerging threats
  • Provides chemical identification for narcotics and hazardous materials detection
 Built for the life you lead
  • Removable rechargeable batteries
  • WiFi connectivity for data sharing
  • Water tight, IP68 rated
  • MIL-STD-810H rated
  • WebUI allows for easy configuration
 Our clear and intuitive interface offers two different modes of operation.
 ID Mode for detailed analysis of unknown chemicals
  • Ideal for advanced users
  • General characterization of chemicals
  • Detailed investigatory functionality
 Screener mode to monitor for the presence of critical chemicals
  • Ideal for all user levels
  • Alerts users to presence of alert substance
  • No interpretation needed to determine next steps
Specifications
Battery TypeRemoveable and Lithium Ion, 7.2V, 49Wh, 4-hr run-time at 25°C
Certifications/ComplianceMIL-STD-810H, IP68 rated
ConnectionsWi-Fi, and USB-C cable
Dimensions (L x W x H)24.1 x 11.7 x 5.6 cm (9.5 x 4.6 x 2.2 in.)
IP RatingIP68
Includes5 MP digital camera, global positioning, WebUI
Laser Excitation Wavelength1064.25 nm ±0.5 nm, 2 cm-1
Laser OutputAdjustable 480 mW, 240 mW, 96 mW
Ocular Hazard Distance160 cm (63 in.)
Temperature Range (Metric)-20°C to 50°C
Weight (English) 3.5 lb.
Weight (Metric)1.6 kg
Unit SizeEach
safety-security-services-278x123
Count on our leading services to maximize your instrument investment.
Learn more ›
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link