(U)HPLC columns optimized for your analytical performance
Solve your separation challenges with our HPLC and UHPLC columns for small molecules. Our popular product lines of Hypersil GOLD and Accucore columns are designed for a wide range of applications.
From the most commonly-used C18 phases, to innovative chemistries, we have the right column to fulfill your chromatography needs
Meet our (U)HPLC column families
Hypersil GOLD columns
Consistency you can rely on, results you can trust
  • Outstanding peak symmetry
  • Excellent lot-to-lot reproducibility
  • Broad range of column formats for sub-2-µm particles
Accucore columns
Resolve your impurities without feeling the pressure
  • Rugged, superficially porous particles
  • Wide selection of chemistries
  • Compatible with HPLC or UHPLC systems
Your first choice of columns for method development
  • Built on 40 years of experience in column development and manufacturing
  • Offered in a complete range of particle sizes, from HPLC to UHPLC, and a wide range of formats, from micro-flow, to analytical, to semi-preparative
  • Able to produce outstanding peak shape, resolution, and lot-to-lot consistency
  • Available with several reversed-phase, HILIC, and ion-exchange phases
 Hypersil GOLD Overview
Explore our complete range of Hypersil GOLD columns to start your chromatography analyses
Hydrophobic Column Phases
Reversed phase chemistry for separation via hydrophobic interactions. Stationary phases with longer carbon chains provide longer retention times and greater hydrophobicity.
C18, C8, C4
Polar-Modified C18 Column Phases
Good for the separation of acidic and basic compounds via reversed phase chromatography using an aqueous mobile phase.
aQ
Phenyl-Based Column Phases
Orthogonal selectivity to traditional hydrophobic phases that utilizes aromatic interactions to achieve challenging separations of aromatic and polar compounds.
Phenyl, PFP
Ion-Exchange Column Phases
Ionic and polar compounds are retained via hydrophilic and ionic interactions with a charged stationary phase. Retained compounds can be eluted by changing the solvent conditions of the column.
Amino, Cyano
Hydrophilic Columns
Ideal for retaining and separating polar compounds based on polarity differences and affinity for a polar stationary phase.
HILIC, Silica
Phase Name
1.9 µm
3 µm
5 µm
Pore Size (Å)
Carbon Load (%)
Surface Area (m²/g)
USP
Hypersil GOLD C18
Checkmark icon
Checkmark icon
Checkmark icon
175
10
220
L1
Hypersil GOLD C8
Checkmark icon
Checkmark icon
Checkmark icon
175
8
220
L7
Hypersil GOLD C4
Checkmark icon
Checkmark icon
Checkmark icon
175
5
220
L26
Hypersil GOLD aQ
Checkmark icon
Checkmark icon
Checkmark icon
175
12
220
L1
Hypersil GOLD PFP
Checkmark icon
Checkmark icon
Checkmark icon
175
8
220
L43
Hypersil GOLD Phenyl
Checkmark icon
Checkmark icon
Checkmark icon
175
8
220
L11
Hypersil GOLD Amino
Checkmark icon
Checkmark icon
Checkmark icon
175
2
220
L9
Hypersil GOLD Cyano
Checkmark icon
Checkmark icon
Checkmark icon
175
4
220
L10
Hypersil GOLD HILIC
Checkmark icon
Checkmark icon
Checkmark icon
175
6
220
—
Hypersil GOLD Silica
Checkmark icon
Checkmark icon
Checkmark icon
175
—
220
L3
Learn more about Hypersil GOLD columns
Solid core silica particles for high-resolution separation at any pressure
Accucore columns offer:
  • UHPLC-level performance, even for HPLC systems with limited pressure capabilities.
  • 14 distinct chemistries to provide a comprehensive tool box for successful (U)HPLC method development.
Our long history of innovation in HPLC stationary phase development,  method development and method transfer provides expertise you can trust.
Accucore HPLC and UHPLC columns
Discover high-throughput columns for rapid (U)HPLC separations
Hydrophobic Column Phases
Reversed phase chemistry for separation via hydrophobic interactions. Stationary phases with longer carbon chains provide longer retention times and greater hydrophobicity.
C18, C8, C4
Specialty Hydrophobic Column PhasesAlternative selectivity to C18 for hydrophobic retention and the geometric separation of structural isomers.
RP-MS, C30
Phenyl-Based Column Phases
Orthogonal selectivity to traditional hydrophobic phases that utilizes aromatic interactions to achieve challenging separations of aromatic and polar compounds.
Biphenyl, Phenyl-Hexyl, PFP
Polar-Modified C18 Column Phases
Good for the separation of acidic and basic compounds via reversed phase chromatography using an aqueous mobile phase
Polar Premium, aQ
HILIC Column Phases
Ideal for retaining and separating polar compounds based on polarity differences and affinity for a polar stationary phase.
HILIC, Amide HILIC
Phase Name
1.5 μm
2.6 μm
4 μm
Pore Size (Å)
Surface Area (m²/g)
Carbon Load (%)
USP
Accucore C18
—
Checkmark icon
—
80
130
9
L1
Accucore Vanquish C18+
Checkmark icon
—
—
80
130
6.5
L1
Accucore XL C18
—
—
Checkmark icon
80
130
7
L1
Accucore C8
—
Checkmark icon
—
80
130
9
L7
Accucore XL C8
—
—
Checkmark icon
80
130
4
L7
Accucore 150-C4
—
Checkmark icon
—
150
80
2
L26
Accucore aQ
—
Checkmark icon
—
80
130
9
L1
Accucore Polar Premium
—
Checkmark icon
—
80
130
8
L60
Accucore RP-MS
—
Checkmark icon
—
80
130
7
L1
Accucore C30
—
Checkmark icon
—
80
130
5
L62
Accucore Biphenyl
—
Checkmark icon
—
80
130
6
L11
Accucore Phenyl-Hexyl
—
Checkmark icon
—
80
130
5
L11
Accucore PFP
—
Checkmark icon
—
80
130
5
L43
Accucore Phenyl-X
—
Checkmark icon
—
80
130
6
–
Accucore HILIC
—
Checkmark icon
—
80
130
–
L3
Accucore 150-Amide-HILIC
—
Checkmark icon
—
150
80
–
–
Accucore Urea HILIC
—
Checkmark icon
—
80
130
–
–
Learn more about Accucore columns
HPLC column selection guide
* Required field
Style Sheet for Global Design System
CMD Wide-format style fixes
Style Sheet for Global Design System
Style Sheet for Komodo Tabs