Product References
Exosomal circRHCG promotes breast cancer metastasis via facilitating M2 polarization through TFEB ubiquitination and degradation.
NPJ precision oncology
Shen HY,Xu JL,Zhang W,Chen QN,Zhu Z,Mao Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Mon Jan 29 00:00:00 UTC 2024
Germline Neurofibromin 1 mutation enhances the anti-tumour immune response and decreases juvenile myelomonocytic leukaemia tumourigenicity.
British journal of haematology
Wang W,Li X,Qin X,Miao Y,Zhang Y,Li S,Yao R,Yang Y,Yu L,Zhu H,Song L,Mao S,Wang X,Chen J,Feng H,Li Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2023
Engineered Human Dendritic Cell Exosomes as Effective Delivery System for Immune Modulation.
International journal of molecular sciences
Elsayed R,Elashiry M,Tran C,Yang T,Carroll A,Liu Y,Hamrick M,Cutler CW
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Tue Jul 11 00:00:00 UTC 2023
USP7 inhibits the progression of nasopharyngeal carcinoma via promoting SPLUNC1-mediated M1 macrophage polarization through TRIM24.
Cell death & disease
Liu H,Tang L,Gong S,Xiao T,Yang H,Gu W,Wang H,Chen P
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Thu Dec 21 00:00:00 UTC 2023
Activation of the JNK/COX-2/HIF-1α axis promotes M1 macrophage via glycolytic shift in HIV-1 infection.
Life science alliance
Zhang J,Yuan Z,Li X,Wang F,Wei X,Kang Y,Mo C,Jiang J,Liang H,Ye L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Uncoupling protein 2 modulates polarization and metabolism of human primary macrophages via glycolysis and the NF‑κB pathway.
Experimental and therapeutic medicine
Lang L,Zheng D,Jiang Q,Meng T,Ma X,Yang Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
WT1 Pulsed Human CD141+ Dendritic Cell Vaccine Has High Potential in Solid Tumor-Targeted Immunotherapy.
International journal of molecular sciences
Cho SY,Jeong SM,Jeon YJ,Yang SJ,Hwang JE,Yoo BM,Kim HS
12-0869-42 was used in Flow cytometry/Cell sorting to develop a DC vaccine named CellgramDC-WT1 (CDW).
Thu Jan 12 00:00:00 UTC 2023
Interleukin 6 exacerbates the progression of warm autoimmune hemolytic anemia by influencing the activity and function of B cells.
Scientific reports
Zhao M,Chen L,Yang J,Zhang Z,Wang H,Shao Z,Liu X,Xing L
12-0869-42 was used in Flow cytometry/Cell sorting to explore the effect of IL-6 on the activity and secretory function of B cells and analyze its effect on clinical indicators and efficacy in wAIHA patients.
Mon Aug 14 00:00:00 UTC 2023
M1 macrophage-derived exosomes and their key molecule lncRNA HOTTIP suppress head and neck squamous cell carcinoma progression by upregulating the TLR5/NF-κB pathway.
Cell death & disease
Jiang H,Zhou L,Shen N,Ning X,Wu D,Jiang K,Huang X
12-0869-42 was used in Flow Cytometry to explore the function of M1-derived exosomes and clarify their specific mechanism in head and neck squamous cell carcinoma (HNSCC).
Thu Feb 24 00:00:00 UTC 2022
M1 macrophage-derived exosomes and their key molecule lncRNA HOTTIP suppress head and neck squamous cell carcinoma progression by upregulating the TLR5/NF-κB pathway.
Cell death & disease
Jiang H,Zhou L,Shen N,Ning X,Wu D,Jiang K,Huang X
12-0869-42 was used in Flow Cytometry to explore the function of M1-derived exosomes and clarify their specific mechanism in head and neck squamous cell carcinoma (HNSCC).
Thu Feb 24 00:00:00 UTC 2022
TcpC Inhibits M1 but Promotes M2 Macrophage Polarization via Regulation of the MAPK/NF-κB and Akt/STAT6 Pathways in Urinary Tract Infection.
Cells
Fang J,Ou Q,Wu B,Li S,Wu M,Qiu J,Cen N,Hu K,Che Y,Ma Y,Pan J
12-0869-42 was used in Flow cytometry/Cell sorting to illuminate the regulatory effects of TcpC on macrophage M1/M2 polarization.
Sun Aug 28 00:00:00 UTC 2022
TcpC Inhibits M1 but Promotes M2 Macrophage Polarization via Regulation of the MAPK/NF-κB and Akt/STAT6 Pathways in Urinary Tract Infection.
Cells
Fang J,Ou Q,Wu B,Li S,Wu M,Qiu J,Cen N,Hu K,Che Y,Ma Y,Pan J
12-0869-42 was used in Flow cytometry/Cell sorting to illuminate the regulatory effects of TcpC on macrophage M1/M2 polarization.
Sun Aug 28 00:00:00 UTC 2022
Glycerol monolaurate inhibition of human B cell activation.
Scientific reports
Fosdick MG,Loftus S,Phillips I,Zacharias ZR,Houtman JCD
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Fri Aug 05 00:00:00 UTC 2022
Longitudinal profiling of respiratory and systemic immune responses reveals myeloid cell-driven lung inflammation in severe COVID-19.
Immunity
Szabo PA,Dogra P,Gray JI,Wells SB,Connors TJ,Weisberg SP,Krupska I,Matsumoto R,Poon MML,Idzikowski E,Morris SE,Pasin C,Yates AJ,Ku A,Chait M,Davis-Porada J,Guo XV,Zhou J,Steinle M,Mackay S,Saqi A,Baldwin MR,Sims PA,Farber DL
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Tue Apr 13 00:00:00 UTC 2021
Small-molecule inhibitors targeting Polycomb repressive complex 1 RING domain.
Nature chemical biology
Shukla S,Ying W,Gray F,Yao Y,Simes ML,Zhao Q,Miao H,Cho HJ,González-Alonso P,Winkler A,Lund G,Purohit T,Kim E,Zhang X,Ray JM,He S,Nikolaidis C,Ndoj J,Wang J,Jaremko Ł,Jaremko M,Ryan RJH,Guzman ML,Grembecka J,Cierpicki T
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
miR-155-5p regulates macrophage M1 polarization and apoptosis in the synovial fluid of patients with knee osteoarthritis.
Experimental and therapeutic medicine
Li GS,Cui L,Wang GD
12-0869 was used in Flow cytometry/Cell sorting to suggest that miR-155-5p may serve as a potential therapeutic target for KOA.
Fri Jan 01 00:00:00 UTC 2021
IL-10-/- Enhances DCs Immunity Against Chlamydia psittaci Infection via OX40L/NLRP3 and IDO/Treg Pathways.
Frontiers in immunology
Li Q,Li X,Quan H,Wang Y,Qu G,Shen Z,He C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Mon Sep 27 00:00:00 UTC 2021
IL-10-/- Enhances DCs Immunity Against Chlamydia psittaci Infection via OX40L/NLRP3 and IDO/Treg Pathways.
Frontiers in immunology
Li Q,Li X,Quan H,Wang Y,Qu G,Shen Z,He C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Mon Sep 27 00:00:00 UTC 2021
Biological and molecular studies on specific immune cells treated with checkpoint inhibitors for the thera-personal approach of breast cancer patients (ex-vivo study).
Oncology research
El-Houseini ME,Arafat MS,El-Husseiny AM,Kasem IM,Kamel MM,El-Habashy AH,Khafagy MM,Radwan EM,Helal MH,Abdellateif MS
12-0869-42 was used in Flow Cytometry to optimize dendritic cell (DC)-based immunotherapy for breast cancer (BC) by co-culturing immune cells (DCs, T cells, tumor-infiltrating lymphocytes, and tumor-infiltrating DCs) treated with anti-PD1 and anti-CTLA4 monoclonal antibodies with autologous breast cancer cells (BCCs), leading to significant upregulation of activation markers on immune cells, downregulation of regulatory T cell markers and key cancer-related genes in BCCs, and changes in protein
Tue Jun 13 00:00:00 UTC 2023
Diffuse large B cell lymphoma-derived extracellular vesicles educate macrophages to promote tumours progression by increasing PGC-1β.
Scandinavian journal of immunology
Liu W,Zhu M,Wang H,Wang W,Lu Y
12-0869-42 was used in Flow Cytometry to signify that the DLBCL-derived EVs mediated the increasing of functional PGC-1β protein in macrophages to promote the tumour progression.
Sat Feb 01 00:00:00 UTC 2020
Breast cancer stem cell RNA-pulsed dendritic cells enhance tumor cell killing by effector T cells.
Oncology letters
Sumransub N,Jirapongwattana N,Jamjuntra P,Thongchot S,Chieochansin T,Yenchitsomanus PT,Thuwajit P,Warnnissorn M,O-Charoenrat P,Thuwajit C
12-0869-42 was used in Flow Cytometry to show the potential use of CSC RNA for priming DCs in modulating an anticancer immune response against BC.
Sun Mar 01 00:00:00 UTC 2020
Antitumor immunity targeting fibroblast activation protein-α in a mouse Lewis lung carcinoma model.
Oncology letters
Xie J,Yuan S,Peng L,Li H,Niu L,Xu H,Guo X,Yang M,Duan F
12-0869 was used in Flow cytometry/Cell sorting to investigate the use of CAFs-targeted immunotherapy to fibroblast activation protein-α (FAP-α) expressed in CAFs.
Wed Jul 01 00:00:00 UTC 2020
Dendritic Cell Maturation Regulates TSPAN7 Function in HIV-1 Transfer to CD4+ T Lymphocytes.
Frontiers in cellular and infection microbiology
Perot BP,García-Paredes V,Luka M,Ménager MM
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Wed Mar 17 00:00:00 UTC 2021
Mesenchymal Stem and Stromal Cells Harness Macrophage-Derived Amphiregulin to Maintain Tissue Homeostasis.
Cell reports
Ko JH,Kim HJ,Jeong HJ,Lee HJ,Oh JY
12-0869-42 was used in Flow Cytometry to demonstrate the role of mesenchymal stem and stromal cell and macrophage cross-talk in post-injury tissue homeostasis in mice.
Tue Mar 17 00:00:00 UTC 2020
The N-Terminal Domain of cGAS Determines Preferential Association with Centromeric DNA and Innate Immune Activation in the Nucleus.
Cell reports
Gentili M,Lahaye X,Nadalin F,Nader GPF,Puig Lombardi E,Herve S,De Silva NS,Rookhuizen DC,Zueva E,Goudot C,Maurin M,Bochnakian A,Amigorena S,Piel M,Fachinetti D,Londoño-Vallejo A,Manel N
12-0869 was used in Flow cytometry/Cell sorting to reveal a preferential functional association of nuclear cGAS with centromeres.
Tue Feb 26 00:00:00 UTC 2019
Reshaping of the Dendritic Cell Chromatin Landscape and Interferon Pathways during HIV Infection.
Cell host & microbe
Johnson JS,Lucas SY,Amon LM,Skelton S,Nazitto R,Carbonetti S,Sather DN,Littman DR,Aderem A
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Wed Mar 14 00:00:00 UTC 2018
A comprehensive characterization of aggravated aging-related changes in T lymphocytes and monocytes in end-stage renal disease: the iESRD study.
Immunity & ageing : I & A
Chiu YL,Shu KH,Yang FJ,Chou TY,Chen PM,Lay FY,Pan SY,Lin CJ,Litjens NHR,Betjes MGH,Bermudez S,Kao KC,Chia JS,Wang G,Peng YS,Chuang YF
Sat Apr 09 00:00:00 UTC 2022
Intrathecal B Cells in MS Have Significantly Greater Lymphangiogenic Potential Compared to B Cells Derived From Non-MS Subjects.
Frontiers in neurology
Stein J,Xu Q,Jackson KC,Romm E,Wuest SC,Kosa P,Wu T,Bielekova B
Wed Sep 30 00:00:00 UTC 2020
AG490 reverses phenotypic alteration of dendritic cells by bladder cancer cells.
Oncology letters
Xiu W,Ma J,Lei T,Zhang M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
Photodynamic therapy reduces the inhibitory effect of osteosarcoma cells on dendritic cells by upregulating HSP70.
Oncology letters
Zhang F,Zhu Y,Fan G,Hu S
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2018
Ginsenoside Rg3 Mitigates Atherosclerosis Progression in Diabetic apoE-/- Mice by Skewing Macrophages to the M2 Phenotype.
Frontiers in pharmacology
Guo M,Xiao J,Sheng X,Zhang X,Tie Y,Wang L,Zhao L,Ji X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Dengue immune sera enhance Zika virus infection in human peripheral blood monocytes through Fc gamma receptors.
PloS one
Li M,Zhao L,Zhang C,Wang X,Hong W,Sun J,Liu R,Yu L,Wang J,Zhang F,Jin X
12-0869 was used in Flow cytometry/Cell sorting to examine the mechanisms of antibody dependent enhancement of Zika virus infection in primary human cells.
Fri Jan 11 00:00:00 UTC 2019
Functionalization-dependent effects of cellulose nanofibrils on tolerogenic mechanisms of human dendritic cells.
International journal of nanomedicine
Tomić S,Ilić N,Kokol V,Gruden-Movsesijan A,Mihajlović D,Bekić M,Sofronić-Milosavljević L,Čolić M,Vučević D
12-0869 was used in Flow cytometry/Cell sorting to investigate how differentially functionalised cellulose nanofibrils affect dendritic cell properties, their viability, morphology, differentiation and maturation potential, and the capacity to regulate T cell-mediated immune response.
Mon Dec 31 00:00:00 UTC 2018
Schistosoma mansoni rSm29 Antigen Induces a Regulatory Phenotype on Dendritic Cells and Lymphocytes From Patients With Cutaneous Leishmaniasis.
Frontiers in immunology
Lopes DM,Oliveira SC,Page B,Carvalho LP,Carvalho EM,Cardoso LS
12-0869 was used in Flow cytometry/Cell sorting to evaluate the potential of the S.mansoni Sm29 antigen to change aspects of profiles of monocyte derived dendritic cells and lymphocytes from subjects with cutaneous leishmaniasis.
Tue Oct 15 00:00:00 UTC 2019
Dexamethasone and Monophosphoryl Lipid A Induce a Distinctive Profile on Monocyte-Derived Dendritic Cells through Transcriptional Modulation of Genes Associated With Essential Processes of the Immune Response.
Frontiers in immunology
García-González PA,Schinnerling K,Sepúlveda-Gutiérrez A,Maggi J,Mehdi AM,Nel HJ,Pesce B,Larrondo ML,Aravena O,Molina MC,Catalán D,Thomas R,Verdugo RA,Aguillón JC
12-0869 was used in Flow cytometry/Cell sorting to study the transcriptional profile of monocyte-derived dendritic cells from healthy subjects to identify molecular regulators and pathways associated with the induction of tolerogenic properties in tolerogenic dendritic cells.
Wed Nov 20 00:00:00 UTC 2019
PD-1 Blockade Promotes Emerging Checkpoint Inhibitors in Enhancing T Cell Responses to Allogeneic Dendritic Cells.
Frontiers in immunology
Stecher C,Battin C,Leitner J,Zettl M,Grabmeier-Pfistershammer K,Höller C,Zlabinger GJ,Steinberger P
Wed Mar 30 00:00:00 UTC 2022
Role of Nuclear Factor (Erythroid-Derived 2)-Like 2 Signaling for Effects of Fumaric Acid Esters on Dendritic Cells.
Frontiers in immunology
Hammer A,Waschbisch A,Knippertz I,Zinser E,Berg J,Jörg S,Kuhbandner K,David C,Pi J,Bayas A,Lee DH,Haghikia A,Gold R,Steinkasserer A,Linker RA
Wed Nov 20 00:00:00 UTC 2019
Dendritic cell maturation, but not type I interferon exposure, restricts infection by HTLV-1, and viral transmission to T-cells.
PLoS pathogens
Rizkallah G,Alais S,Futsch N,Tanaka Y,Journo C,Mahieux R,Dutartre H
12-0869 was used in Flow cytometry/Cell sorting to conclude that the nature of the DCs encountered by HTLV-1 upon primo-infection and the viral trafficking route through the vesicular pathway of these cells determine the efficiency of viral transmission to T-cells, which may condition the fate of infection.
Sat Apr 01 00:00:00 UTC 2017
Modelling IRF8 Deficient Human Hematopoiesis and Dendritic Cell Development with Engineered iPS Cells.
Stem cells (Dayton, Ohio)
Sontag S,Förster M,Qin J,Wanek P,Mitzka S,Schüler HM,Koschmieder S,Rose-John S,Seré K,Zenke M
12-0869 was used in Flow cytometry/Cell sorting to engineer a human IRF8 knockout model that allows studying molecular mechanisms of human immunodeficiencies in vitro, including the pathophysiology of IRF8 deficient DC.
Sat Apr 01 00:00:00 UTC 2017
Abnormal phenotypic features of IgM+B cell subsets in patients with chronic hepatitis C virus infection.
Experimental and therapeutic medicine
Kong F,Feng B,Zhang H,Rao H,Wang J,Cong X,Wei L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2017
MALP-2, an agonist of TLR6, promotes the immune status without affecting the differentiation capacity of umbilical cord mesenchymal stem cells.
Experimental and therapeutic medicine
Wu X,Xu L,Shen Y,Yu N,Zhang Y,Guo T
12-0869 was used in Flow cytometry/Cell sorting to assess the role of TLR6 in the immune status of umbilical cord mesenchymal stem cells, by stimulation with the TLR6 agonist MALP-2.
Fri Dec 01 00:00:00 UTC 2017
Allostimulatory capacity of conditionally immortalized proximal tubule cell lines for bioartificial kidney application.
Scientific reports
Mihajlovic M,van den Heuvel LP,Hoenderop JG,Jansen J,Wilmer MJ,Westheim AJF,Allebes WA,Stamatialis D,Hilbrands LB,Masereeuw R
12-0869 was used in Flow cytometry/Cell sorting to assess the immunosafety of conditionally immortalized proximal tubule cell lines for use in bioartificial kidneys, specifically by investigating their allogeneic efects.
Wed Aug 02 00:00:00 UTC 2017
Pharmacological Inhibition of the Histone Lysine Demethylase KDM1A Suppresses the Growth of Multiple Acute Myeloid Leukemia Subtypes.
Cancer research
McGrath JP,Williamson KE,Balasubramanian S,Odate S,Arora S,Hatton C,Edwards TM,O'Brien T,Magnuson S,Stokoe D,Daniels DL,Bryant BM,Trojer P
12-0869 was used in Flow cytometry/Cell sorting to investigate the consequences of inhibiting lysine-specific demethylase 1 in a panel of cell lines representing all acute myelogenous leukaemia subtypes.
Fri Apr 01 00:00:00 UTC 2016
MicroRNA-720 suppresses M2 macrophage polarization by targeting GATA3.
Bioscience reports
Zhong Y,Yi C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 12-0869-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
An in vitro model of granuloma-like cell aggregates substantiates early host immune responses against Mycobacterium massiliense infection.
Biology open
Je S,Quan H,Na Y,Cho SN,Kim BJ,Seok SH
12-0869 was used in Flow cytometry/Cell sorting to study the interaction between host cells and Mycobacterium massiliense.
Mon Aug 15 00:00:00 UTC 2016
Pentraxin-3 regulates the inflammatory activity of macrophages.
Biochemistry and biophysics reports
Shiraki A,Kotooka N,Komoda H,Hirase T,Oyama JI,Node K
12-0869 was used in Immunocytochemistry to investigate the function of pentraxin-3 in macrophages in vitro.
Tue Mar 01 00:00:00 UTC 2016
Antibodies to P-selectin glycoprotein ligand-1 block dendritic cell-mediated enterovirus 71 transmission and prevent virus-induced cells death.
Virulence
Ren XX,Li C,Xiong SD,Huang Z,Wang JH,Wang HB
12-0869 was used in Flow cytometry/Cell sorting to demonstrate that PSGL-1 present on the cell surfaces of monocyte-derived dendritic cells mediates enterovirus 71 binding and facilitates further infection.
Thu Oct 06 00:00:00 UTC 2016
The role of Toll-like receptor 3 and 4 in regulating the function of mesenchymal stem cells isolated from umbilical cord.
International journal of molecular medicine
Zhang L,Liu D,Pu D,Wang Y,Li L,He Y,Li Y,Li L,Qiu Z,Zhao S,Li W
12-0869-42 was used in Flow Cytometry to examine the role of the TLR3 and TLR4 pathways in modulating the immune properties of human umbilical cord-derived MSCs.
Wed Apr 01 00:00:00 UTC 2015
CX₃CR1⁺ mononuclear phagocytes support colitis-associated innate lymphoid cell production of IL-22.
The Journal of experimental medicine
Longman RS,Diehl GE,Victorio DA,Huh JR,Galan C,Miraldi ER,Swaminath A,Bonneau R,Scherl EJ,Littman DR
12-0869 was used in Flow cytometry/Cell sorting to study the role of CX3CR1+ mononuclear phagocytes in the development of colitis and disease progression following Citrobacter rodentium infection.
Mon Jul 28 00:00:00 UTC 2014
M-CSF inhibits anti-HIV-1 activity of IL-32, but they enhance M2-like phenotypes of macrophages.
Journal of immunology (Baltimore, Md. : 1950)
Osman A,Bhuyan F,Hashimoto M,Nasser H,Maekawa T,Suzu S
12-0869 was used in Flow cytometry/Cell sorting to investigate the functional interplay between M-CSF and IL-32 acting on macrophages.
Sun Jun 01 00:00:00 UTC 2014
Dendritic cell profile induced by Schistosoma mansoni antigen in cutaneous leishmaniasis patients.
BioMed research international
Lopes DM,Fernandes JS,Cardoso TM,Bafica AM,Oliveira SC,Carvalho EM,Araujo MI,Cardoso LS
12-0869 was used in Flow cytometry/Cell sorting to investigate the dendritic cell profile induced by Schistosoma mansoni antigen in cutaneous leishmaniasis patients.
Thu Jul 02 00:00:00 UTC 2015
Interferon beta and vitamin D synergize to induce immunoregulatory receptors on peripheral blood monocytes of multiple sclerosis patients.
PloS one
Waschbisch A,Sanderson N,Krumbholz M,Vlad G,Theil D,Schwab S,Mäurer M,Derfuss T
12-0869 was used in Flow cytometry/Cell sorting to investigate whether the immunoregulatory receptors immunoglobulin-like transcript 3 and 4 mediate the therapeutic effect of interferon beta in multiple sclerosis.
Tue Sep 08 00:00:00 UTC 2015
Brugia malayi microfilariae induce a regulatory monocyte/macrophage phenotype that suppresses innate and adaptive immune responses.
PLoS neglected tropical diseases
O'Regan NL,Steinfelder S,Venugopal G,Rao GB,Lucius R,Srikantam A,Hartmann S
12-0869 was used in Flow cytometry/Cell sorting to show that monocytes stimulated with Brugia malayi microfilarial lysate develop a defined regulatory phenotype.
Wed Oct 01 00:00:00 UTC 2014
Tumor necrosis factor receptor 1 associates with CD137 ligand and mediates its reverse signaling.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Moh MC,Lorenzini PA,Gullo C,Schwarz H
12-0869 was used in Flow cytometry/Cell sorting to determine the mechanisms behind the activation of monocytes by reverse signalling through the CD137 ligand.
Thu Aug 01 00:00:00 UTC 2013
Leishmania major infection in humanized mice induces systemic infection and provokes a nonprotective human immune response.
PLoS neglected tropical diseases
Wege AK,Florian C,Ernst W,Zimara N,Schleicher U,Hanses F,Schmid M,Ritter U
12-0869 was used in Flow cytometry/Cell sorting to describe Leishmania major infection in humanised mice for the first time, characterising disease development and the induction of the human immune response.
Thu Nov 01 00:00:00 UTC 2012
A Francisella tularensis live vaccine strain that improves stimulation of antigen-presenting cells does not enhance vaccine efficacy.
PloS one
Schmitt DM,O'Dee DM,Horzempa J,Carlson PE,Russo BC,Bales JM,Brown MJ,Nau GJ
12-0869 was used in Flow cytometry/Cell sorting to examine whether the interaction of genetically modified Francisella tularensis live vaccine strains with human antigen-presenting cells correlates with vaccine effectiveness.
Mon Jun 18 00:00:00 UTC 2012
Vitamin D suppression of endoplasmic reticulum stress promotes an antiatherogenic monocyte/macrophage phenotype in type 2 diabetic patients.
The Journal of biological chemistry
Riek AE,Oh J,Sprague JE,Timpson A,de las Fuentes L,Bernal-Mizrachi L,Schechtman KB,Bernal-Mizrachi C
12-0869 was used in Flow cytometry/Cell sorting to investigate the the possible role of vitamin D deficiency as a mechanism of increased atherosclerosis in T2DM patients.
Fri Nov 09 00:00:00 UTC 2012
Optimal oligonucleotide sequences for TLR9 inhibitory activity in human cells: lack of correlation with TLR9 binding.
International immunology
Ashman RF,Goeken JA,Latz E,Lenert P
12-0869 was used in Flow cytometry/Cell sorting to explore the use of oligonucleotide sequences for therapeutic purposes in situations where inflammation is enhanced by TLR9.
Tue Mar 01 00:00:00 UTC 2011
Penicillium marneffei-stimulated dendritic cells enhance HIV-1 trans-infection and promote viral infection by activating primary CD4+ T cells.
PloS one
Qin Y,Li Y,Liu W,Tian R,Guo Q,Li S,Li H,Zhang D,Zheng Y,Wu L,Lan K,Wang J
12-0869 was used in Flow cytometry/Cell sorting to demonstrate that DC interaction with HIV-1 has been exploited by P. marneffei for viral amplification.
Mon Mar 26 00:00:00 UTC 2012
Dendritic cells express hematopoietic prostaglandin D synthase and function as a source of prostaglandin D2 in the skin.
The American journal of pathology
Shimura C,Satoh T,Igawa K,Aritake K,Urade Y,Nakamura M,Yokozeki H
12-0869 was used in Flow cytometry/Cell sorting to determine that dendritic cells may mediate or regulate skin inflammation by releasing PGD2 in response to various stimuli.
Fri Jan 01 00:00:00 UTC 2010
Novel role of plasmacytoid dendritic cells in humans: induction of interleukin-10-producing Treg cells by plasmacytoid dendritic cells in patients with rheumatoid arthritis responding to therapy.
Arthritis and rheumatism
Kavousanaki M,Makrigiannakis A,Boumpas D,Verginis P
12-0869 was used in Flow cytometry/Cell sorting to explore the contribution of dendritic cells to the development of regulatory T cells in a human autoimmune disease setting.
Fri Jan 01 00:00:00 UTC 2010
Immunological aspects of REIC/Dkk-3 in monocyte differentiation and tumor regression.
International journal of oncology
Watanabe M,Kashiwakura Y,Huang P,Ochiai K,Futami J,Li SA,Takaoka M,Nasu Y,Sakaguchi M,Huh NH,Kumon H
12-0869-42 was used in Flow cytometry/Cell sorting to study that recombinant REIC/Dkk-3 protein induced the differentiation of human CD14+ monocytes into a novel cell type.
Sun Mar 01 00:00:00 UTC 2009
Polychromatic flow cytometric high-throughput assay to analyze the innate immune response to Toll-like receptor stimulation.
Journal of immunological methods
Jansen K,Blimkie D,Furlong J,Hajjar A,Rein-Weston A,Crabtree J,Reikie B,Wilson C,Kollmann T
12-0869 was used in Flow cytometry/Cell sorting to describe a novel platform that enables the interrogation and screening of APC responses to TLR ligands.
Thu Jul 31 00:00:00 UTC 2008
Enhanced efficacy of CTLA-4 fusion anti-caries DNA vaccines in gnotobiotic hamsters.
Acta pharmacologica Sinica
Zhang F,Li YH,Fan MW,Jia R,Xu QA,Guo JH,Yu F,Tian QW
12-0869 was used in Flow cytometry/Cell sorting to prove CTLLA-4-antigen fusion enhances anti-caries DNA vaccines.
Wed Aug 01 00:00:00 UTC 2007
Clinical significance of costimulatory molecules CD80/CD86 expression in IgA nephropathy.
Kidney international
Wu Q,Jinde K,Endoh M,Sakai H
12-0869 was used in Flow cytometry/Cell sorting to suggest that CD80 and CD86 activate T cells in IgAN, CD80/CD86 expressions correlated with renal function at the time of renal biopsy, and monocyte/macrophages and tubular epithelial cells act as APC.
Mon Mar 01 00:00:00 UTC 2004