    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • TIM-3 Antibodies
            Invitrogen
            CD366 (TIM3) Monoclonal Antibody (F38-2E2), PE, eBioscience™
            4 Published Figures
            15 References
            View all (143) TIM-3 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 12-3109-41
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            Price (USD)
            141.95
            Special offer
            Online exclusive
            167.00
            Save 25.05 (15%)
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers
            Cite CD366 (TIM3) Monoclonal Antibody (F38-2E2), PE, eBioscience™
            • Antibody Testing Data (1)
            • Published Figures (4)
            Application
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.
            FIGURE: 1 / 5
            CD366 (TIM3) Antibody (12-3109-41) in Flow
            Staining of 3-day Anti-Human CD3 and CD28 Functional Grade Purifieds (Product # 16-0039-81 and Product # 16-0289-81)-stimulated normal human peripheral blood cells with Anti-Human CD4 FITC (Product # 11-0049-42) and Mouse IgG1 K Isotype Control PE (Product # 12-4714-81) (left) or Anti-Human CD366 (TIM3) PE (right). Total viable cells, as determined by Fixable Viability Dye eFluor® 660 (Product # 65-0864-14), were used for analysis.
            View Product
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            CD366 (TIM3) Antibody in Flow Cytometry (Flow)
            Product Details
            12-3109-41
            Applications
            Tested Dilution
            Publications
            Flow Cytometry (Flow)
            5 µL (0.125 µg)/test
            View 15 publications 15 publications
            Product Specifications
            Species Reactivity
            Human
            Published species
            Human, Mouse
            Host/Isotype
            Mouse / IgG1, kappa
            Recommended Isotype Control
            Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PE, eBioscience™
            Class
            Monoclonal
            Type
            Antibody
            Clone
            F38-2E2
            Conjugate
            PE
            • Alexa Fluor 532 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Brilliant UV 805
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 786
            • FITC
            • Functional Grade
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation
            Excitation/Emission Max
            565/576 nm View spectra spectra
            Form
            Liquid
            Concentration
            5 µL/Test
            Purification
            Affinity chromatography
            Storage buffer
            PBS, pH 7.2, with BSA
            Contains
            0.09% sodium azide
            Storage conditions
            4°C, store in dark, DO NOT FREEZE!
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            RRID
            AB_2572604
            Product Specific Information
            Description: This F38-2E2 monoclonal antibody reacts with human CD366, also known as T cell immunoglobulin and mucin domain-containing protein 3 (TIM3) or HAVCR2. This cell surface receptor is expressed on activated CD4+ T cell subsets (e.g. Th1, Th17, and Treg), CD8+ T cells, monocytes, dendritic cells, and mast cells. Due to alternative splicing, CD366 exists as membrane-bound and soluble forms. Galectin-9 has been identified as the ligand for CD366. In humans, this receptor negatively regulates CD4+ T cells, influencing the secretion of some Th1- and Th17-related cytokines. CD366 has also been implicated in tolerance, autoimmune disease (e.g. multiple sclerosis), and HIV infection.
            Applications Reported: This F38-2E2 antibody has been reported for use in flow cytometric analysis.
            Applications Tested: This F38-2E2 antibody has been pre-titrated and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
            Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
            Filtration: 0.2 µm post-manufacturing filtered.
            Target Information
            TIM3 (Hepatitis A virus cellular receptor 2, HAVCR2, T-cell immunoglobulin, mucin-dmain containing-3) is a 281 amino acid long, Type-1 Th1- specific cell surface glycoprotein expressed on terminally differentiated CD4+Th1 and CD8+Tc1 cells. TIM3 consists of an IgV-like domain, a mucin-like domain in the extracellular region, and a conserved Tyrosine phosphorylation motif in the cytoplasmic region. TIM3 is involved in macrophage activation and induction of autoimmune diseases. Further, TIM3 down-regulates aggressive Th1-mediated immune responses and facilitates in the development of immune tolerance. Pathological significance of TIM3 has been attributed to Experimental autoimmune encephalomyelitis (EAE), a Th-1 dependent autoimmune disease, and also enhances the severity of experimental autoimmune encephalomyelitis in mice.
            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 8:21
            Loaded: 0.00%
            0:00
            Stream Type LIVE
            Remaining Time -8:21
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected
            • en (Main), selected
            This is a modal window.
            Beginning of dialog window. Escape will cancel and close the window.
            Text
            Text Background
            Caption Area Background
            Font Size
            Text Edge Style
            Font Family
            End of dialog window.
            This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
            How to use the Panel Builder
            Bioinformatics
            Protein Aliases: CD366; FLJ14428; HAVcr-2; Hepatitis A virus cellular receptor 2; kidney injury molecule-3; sCD366; soluble CD366; soluble TIM 3; T cell immunoglobulin mucin 3; T cell immunoglobulin mucin-3; T-cell immunoglobulin and mucin domain-containing protein 3; T-cell immunoglobulin mucin family member 3; T-cell immunoglobulin mucin receptor 3; T-cell membrane protein 3; TIM-3; TIMD-3
            View more View less
            Gene Aliases: CD366; HAVcr-2; HAVCR2; KIM-3; Tim-3; TIM3; TIMD-3; TIMD3
            View more View less
            UniProt ID: (Human) Q8TDQ0
            View more View less
            Entrez Gene ID: (Human) 84868
            View more View less
            Function(s)
            protein binding metal ion binding immunoglobulin receptor superfamily
            Process(es)
            adaptive immune response macrophage activation involved in immune response natural killer cell tolerance induction regulation of tolerance induction dependent upon immune response negative regulation of T-helper 1 type immune response negative regulation of natural killer cell mediated cytotoxicity directed against tumor cell target inflammatory response negative regulation of gene expression negative regulation of myeloid dendritic cell activation negative regulation of NF-kappaB transcription factor activity negative regulation of interferon-alpha production negative regulation of interferon-gamma production negative regulation of interleukin-2 production negative regulation of interleukin-3 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production positive regulation of chemokine production positive regulation of interferon-gamma production positive regulation of interleukin-1 production positive regulation of interleukin-4 production negative regulation of natural killer cell activation toll-like receptor 3 signaling pathway toll-like receptor 7 signaling pathway toll-like receptor 9 signaling pathway positive regulation of T cell proliferation negative regulation of T cell proliferation positive regulation of macrophage activation innate immune response defense response to Gram-positive bacterium maternal process involved in female pregnancy positive regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide negative regulation of granulocyte colony-stimulating factor production negative regulation of defense response to bacterium positive regulation of defense response to bacterium positive regulation of NIK/NF-kappaB signaling positive regulation of tumor necrosis factor secretion negative regulation of immunological synapse formation negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-7c4bf66445-pxmrt:80/100.66.79.171:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0