Product References
IL-6 signaling drives self-renewal and alternative activation of adipose tissue macrophages.
Frontiers in immunology
Ackermann J,Arndt L,Fröba J,Lindhorst A,Glaß M,Kirstein M,Hobusch C,Wunderlich FT,Braune J,Gericke M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Mar 15 00:00:00 UTC 2024
TL1A is an epithelial alarmin that cooperates with IL-33 for initiation of allergic airway inflammation.
The Journal of experimental medicine
Schmitt P,Duval A,Camus M,Lefrançais E,Roga S,Dedieu C,Ortega N,Bellard E,Mirey E,Mouton-Barbosa E,Burlet-Schiltz O,Gonzalez-de-Peredo A,Cayrol C,Girard JP
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Jun 03 00:00:00 UTC 2024
Spleen-targeted neoantigen DNA vaccine for personalized immunotherapy of hepatocellular carcinoma.
EMBO molecular medicine
Wu M,Luo Z,Cai Z,Mao Q,Li Z,Li H,Zhang C,Zhang Y,Zhong A,Wu L,Liu X
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Oct 11 00:00:00 UTC 2023
The miR-23-27-24 clusters drive lipid-associated macrophage proliferation in obese adipose tissue.
Cell reports
Sprenkle NT,Winn NC,Bunn KE,Zhao Y,Park DJ,Giese BG,Karijolich JJ,Ansel KM,Serezani CH,Hasty AH,Pua HH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Aug 29 00:00:00 UTC 2023
Involvement of langerin in the protective function of a keratan sulfate-based disaccharide in an emphysema mouse model.
The Journal of biological chemistry
Ohkawa Y,Kanto N,Nakano M,Fujinawa R,Kizuka Y,Johnson EL,Harada Y,Tamura JI,Taniguchi N
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Oncolytic virus M1 functions as a bifunctional checkpoint inhibitor to enhance the antitumor activity of DC vaccine.
Cell reports. Medicine
Dan J,Cai J,Zhong Y,Wang C,Huang S,Zeng Y,Fan Z,Xu C,Hu L,Zhang J,Hu J,Liu Y,Su X,Zhu W,Yan G,Liang J,Lin Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Oct 17 00:00:00 UTC 2023
Guidelines for mouse and human DC generation.
European journal of immunology
Lutz MB,Ali S,Audiger C,Autenrieth SE,Berod L,Bigley V,Cyran L,Dalod M,Dörrie J,Dudziak D,Flórez-Grau G,Giusiano L,Godoy GJ,Heuer M,Krug AB,Lehmann CHK,Mayer CT,Naik SH,Scheu S,Schreibelt G,Segura E,Seré K,Sparwasser T,Tel J,Xu H,Zenke M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2023
Engineered oncolytic bacteria HCS1 exerts high immune stimulation and safety profiles for cancer therapy.
Theranostics
Sun Y,Guo Y,Liu X,Liu J,Sun H,Li Z,Wen M,Jiang SN,Tan W,Zheng JH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Nov 02 00:00:00 UTC 2023
Oral Immunotherapy Reshapes Intestinal Immunosuppression via Metabolic Reprogramming to Enhance Systemic Anti-Tumor Immunity.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Cao X,Xu Y,Zhou C,Huo J,Su S,Liu L,Zhu Z,Li L,Jia W,Wang C,Zhen M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Toxin expression during Staphylococcus aureus infection imprints host immunity to inhibit vaccine efficacy.
NPJ vaccines
Teymournejad O,Li Z,Beesetty P,Yang C,Montgomery CP
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Jan 24 00:00:00 UTC 2023
Topical Diacerein Decreases Skin and Splenic CD11c+ Dendritic Cells in Psoriasis.
International journal of molecular sciences
Brunner SM,Ramspacher A,Rieser C,Leitner J,Heil H,Ablinger M,Tevini J,Wimmer M,Koller A,Piñón Hofbauer J,Felder TK,Bauer JW,Kofler B,Lang R,Wally V
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Feb 21 00:00:00 UTC 2023
Outer Membrane Vesicle-Coated Nanoparticle Vaccine Protects Against Acinetobacter baumannii Pneumonia and Sepsis.
Advanced nanobiomed research
Bjanes E,Zhou J,Qayum T,Krishnan N,Zurich RH,Menon ND,Hoffman A,Fang RH,Zhang L,Nizet V
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2023
CD11c+ microglia promote white matter repair after ischemic stroke.
Cell death & disease
Jia J,Zheng L,Ye L,Chen J,Shu S,Xu S,Bao X,Xia S,Liu R,Xu Y,Zhang M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Feb 24 00:00:00 UTC 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
P. gingivalis Infection Upregulates PD-L1 Expression on Dendritic Cells, Suppresses CD8+ T-cell Responses, and Aggravates Oral Cancer.
Cancer immunology research
Ren J,Han X,Lohner H,Hoyle RG,Li J,Liang S,Wang H
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Inhibition of Macrophage-Specific CHIT1 as an Approach to Treat Airway Remodeling in Severe Asthma.
International journal of molecular sciences
Sklepkiewicz P,Dymek B,Mlacki M,Zagozdzon A,Salamon M,Siwińska AM,Mazurkiewicz MP,de Souza Xavier Costa N,Mazur M,Mauad T,Gołębiowski A,Dzwonek K,Gołąb J,Zasłona Z
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Targeted inhibition of osteoclastogenesis reveals the pathogenesis and therapeutics of bone loss under sympathetic neurostress.
International journal of oral science
Sui B,Liu J,Zheng C,Dang L,Chen J,Cao Y,Zhang K,Liu L,Dang M,Zhang L,Chen N,He T,Xuan K,Jin F,Zhang G,Jin Y,Hu C
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Mon Aug 01 00:00:00 UTC 2022
Meningeal lymphatic drainage promotes T cell responses against Toxoplasma gondii but is dispensable for parasite control in the brain.
eLife
Kovacs MA,Cowan MN,Babcock IW,Sibley LA,Still K,Batista SJ,Labuzan SA,Sethi I,Harris TH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2022
Interferon-γ Stimulates Interleukin-27 Derived from Dendritic Cells to Regulate Th9 Differentiation through STAT1/3 Pathway.
Disease markers
Xiong P,Liu T,Chen Y,Xu Q,Hu X,Han F,Shi L
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Oct 31 00:00:00 UTC 2022
Neutrophil-dendritic cell interaction plays an important role in live attenuated Leishmania vaccine induced immunity.
PLoS neglected tropical diseases
Bhattacharya P,Ismail N,Saxena A,Gannavaram S,Dey R,Oljuskin T,Akue A,Takeda K,Yu J,Karmakar S,Dagur PK,McCoy JP,Nakhasi HL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Feb 01 00:00:00 UTC 2022
Self-Tolerance of Vascular Tissues Is Broken Down by Vascular Dendritic Cells in Response to Systemic Inflammation to Initiate Regional Autoinflammation.
Frontiers in immunology
Sun L,Zhang W,Zhao L,Zhao Y,Wang F,Lew AM,Xu Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Fri Apr 01 00:00:00 UTC 2022
Nucleolin Therapeutic Targeting Decreases Pancreatic Cancer Immunosuppression.
Cancers
Ponzo M,Debesset A,Cossutta M,Chalabi-Dchar M,Houppe C,Pilon C,Nicolas-Boluda A,Meunier S,Raineri F,Thiolat A,Nicolle R,Maione F,Brundu S,Cojocaru CF,Bouvet P,Bousquet C,Gazeau F,Tournigand C,Courty J,Giraudo E,Cohen JL,Cascone I
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Aug 31 00:00:00 UTC 2022
Single-cell transcriptomics reveals distinct cell response between acute and chronic pulmonary infection of Pseudomonas aeruginosa.
MedComm
Hu X,Wu M,Ma T,Zhang Y,Zou C,Wang R,Zhang Y,Ren Y,Li Q,Liu H,Li H,Wang T,Sun X,Yang Y,Tang M,Li X,Li J,Gao X,Li T,Zhou X
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Thu Dec 01 00:00:00 UTC 2022
Neutrophil-specific gain-of-function mutations in Nlrp3 promote development of cryopyrin-associated periodic syndrome.
The Journal of experimental medicine
Stackowicz J,Gaudenzio N,Serhan N,Conde E,Godon O,Marichal T,Starkl P,Balbino B,Roers A,Bruhns P,Jönsson F,Moguelet P,Georgin-Lavialle S,Broderick L,Hoffman HM,Galli SJ,Reber LL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Mon Oct 04 00:00:00 UTC 2021
Loss of p53 in mesenchymal stem cells promotes alteration of bone remodeling through negative regulation of osteoprotegerin.
Cell death and differentiation
Velletri T,Huang Y,Wang Y,Li Q,Hu M,Xie N,Yang Q,Chen X,Chen Q,Shou P,Gan Y,Candi E,Annicchiarico-Petruzzelli M,Agostini M,Yang H,Melino G,Shi Y,Wang Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Depletion of Adipocyte Becn1 Leads to Lipodystrophy and Metabolic Dysregulation.
Diabetes
Jin Y,Ji Y,Song Y,Choe SS,Jeon YG,Na H,Nam TW,Kim HJ,Nahmgoong H,Kim SM,Kim JW,Nam KT,Seong JK,Hwang D,Park CB,Lee IH,Kim JB,Lee HW
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Depletion of Adipocyte Becn1 Leads to Lipodystrophy and Metabolic Dysregulation.
Diabetes
Jin Y,Ji Y,Song Y,Choe SS,Jeon YG,Na H,Nam TW,Kim HJ,Nahmgoong H,Kim SM,Kim JW,Nam KT,Seong JK,Hwang D,Park CB,Lee IH,Kim JB,Lee HW
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Induction of IL-12p40 and type 1 immunity by Toxoplasma gondii in the absence of the TLR-MyD88 signaling cascade.
PLoS pathogens
Snyder LM,Doherty CM,Mercer HL,Denkers EY
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Oct 01 00:00:00 UTC 2021
Pancreatic Cancer Chemotherapy Is Potentiated by Induction of Tertiary Lymphoid Structures in Mice.
Cellular and molecular gastroenterology and hepatology
Delvecchio FR,Fincham REA,Spear S,Clear A,Roy-Luzarraga M,Balkwill FR,Gribben JG,Bombardieri M,Hodivala-Dilke K,Capasso M,Kocher HM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Mar 28 00:00:00 UTC 2022
T cell-derived tumor necrosis factor induces cytotoxicity by activating RIPK1-dependent target cell death.
JCI insight
Chun N,Ang RL,Chan M,Fairchild RL,Baldwin WM,Horwitz JK,Gelles JD,Chipuk JE,Kelliher MA,Pavlov VI,Li Y,Homann D,Heeger PS,Ting AT
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Dec 22 00:00:00 UTC 2021
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Jan 24 00:00:00 UTC 2022
Enrichment of Neurodegenerative Microglia Signature in Brain-Derived Extracellular Vesicles Isolated from Alzheimer's Disease Mouse Models.
Journal of proteome research
Muraoka S,Jedrychowski MP,Iwahara N,Abdullah M,Onos KD,Keezer KJ,Hu J,Ikezu S,Howell GR,Gygi SP,Ikezu T
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Western Blot
Fri Mar 05 00:00:00 UTC 2021
Intratumoral injection of TLR9 agonist promotes an immunopermissive microenvironment transition and causes cooperative antitumor activity in combination with anti-PD1 in pancreatic cancer.
Journal for immunotherapy of cancer
Carbone C,Piro G,Agostini A,Delfino P,De Sanctis F,Nasca V,Spallotta F,Sette C,Martini M,Ugel S,Corbo V,Cappello P,Bria E,Scarpa A,Tortora G
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Sep 01 00:00:00 UTC 2021
The Intermucosal Connection between the Mouth and Gut in Commensal Pathobiont-Driven Colitis.
Cell
Kitamoto S,Nagao-Kitamoto H,Jiao Y,Gillilland MG,Hayashi A,Imai J,Sugihara K,Miyoshi M,Brazil JC,Kuffa P,Hill BD,Rizvi SM,Wen F,Bishu S,Inohara N,Eaton KA,Nusrat A,Lei YL,Giannobile WV,Kamada N
13-0114-82 was used in Flow Cytometry to conclude that oral inflammation, such as periodontitis, exacerbates gut inflammation by supplying the gut with both colitogenic pathobionts and pathogenic T cells.
Thu Jul 23 00:00:00 UTC 2020
Dual-Wavelength Photosensitive Nano-in-Micro Scaffold Regulates Innate and Adaptive Immune Responses for Osteogenesis.
Nano-micro letters
Zhao Q,Shi M,Yin C,Zhao Z,Zhang J,Wang J,Shen K,Zhang L,Tang H,Xiao Y,Zhang Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sat Nov 21 00:00:00 UTC 2020
Dual-Wavelength Photosensitive Nano-in-Micro Scaffold Regulates Innate and Adaptive Immune Responses for Osteogenesis.
Nano-micro letters
Zhao Q,Shi M,Yin C,Zhao Z,Zhang J,Wang J,Shen K,Zhang L,Tang H,Xiao Y,Zhang Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sat Nov 21 00:00:00 UTC 2020
Glufosinate constrains synchronous and metachronous metastasis by promoting anti-tumor macrophages.
EMBO molecular medicine
Menga A,Serra M,Todisco S,Riera-Domingo C,Ammarah U,Ehling M,Palmieri EM,Di Noia MA,Gissi R,Favia M,Pierri CL,Porporato PE,Castegna A,Mazzone M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Wed Oct 07 00:00:00 UTC 2020
Glufosinate constrains synchronous and metachronous metastasis by promoting anti-tumor macrophages.
EMBO molecular medicine
Menga A,Serra M,Todisco S,Riera-Domingo C,Ammarah U,Ehling M,Palmieri EM,Di Noia MA,Gissi R,Favia M,Pierri CL,Porporato PE,Castegna A,Mazzone M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Wed Oct 07 00:00:00 UTC 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Notch2 complements Notch1 to mediate inductive signaling that initiates early T cell development.
The Journal of cell biology
Romero-Wolf M,Shin B,Zhou W,Koizumi M,Rothenberg EV,Hosokawa H
13-0114 was used in Flow cytometry/Cell sorting to define a distinct mechanism of Notch signal response that distinguishes the initial stages of murine T cell development.
Mon Oct 05 00:00:00 UTC 2020
RIG-I Promotes Cell Death in Hepatocellular Carcinoma by Inducing M1 Polarization of Perineal Macrophages Through the RIG-I/MAVS/NF-κB Pathway.
OncoTargets and therapy
Zhou B,Li C,Yang Y,Wang Z
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Apr 17 00:00:00 UTC 2022
Diversity in medullary thymic epithelial cells controls the activity and availability of iNKT cells.
Nature communications
Lucas B,White AJ,Cosway EJ,Parnell SM,James KD,Jones ND,Ohigashi I,Takahama Y,Jenkinson WE,Anderson G
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon May 04 00:00:00 UTC 2020
Analysis of the immune response to sciatic nerve injury identifies efferocytosis as a key mechanism of nerve debridement.
eLife
Kalinski AL,Yoon C,Huffman LD,Duncker PC,Kohen R,Passino R,Hafner H,Johnson C,Kawaguchi R,Carbajal KS,Jara JS,Hollis E,Geschwind DH,Segal BM,Giger RJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Dec 02 00:00:00 UTC 2020
CD103+ cDC1 and endogenous CD8+ T cells are necessary for improved CD40L-overexpressing CAR T cell antitumor function.
Nature communications
Kuhn NF,Lopez AV,Li X,Cai W,Daniyan AF,Brentjens RJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Dec 02 00:00:00 UTC 2020
Ovarian granulosa cell tumor characterization identifies FOXL2 as an immunotherapeutic target.
JCI insight
Pierini S,Tanyi JL,Simpkins F,George E,Uribe-Herranz M,Drapkin R,Burger R,Morgan MA,Facciabene A
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Aug 20 00:00:00 UTC 2020
Hepatocytes trap and silence coxsackieviruses, protecting against systemic disease in mice.
Communications biology
Kimura T,Flynn CT,Whitton JL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Oct 16 00:00:00 UTC 2020
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jun 17 00:00:00 UTC 2021
Zdhhc2 Is Essential for Plasmacytoid Dendritic Cells Mediated Inflammatory Response in Psoriasis.
Frontiers in immunology
Zhou B,Yang W,Li W,He L,Lu L,Zhang L,Liu Z,Wang Y,Chao T,Huang R,Gu Y,Jia T,Liu Q,Tian S,Pierre P,Maeda T,Liang Y,Kong E
13-0114-85 was used in Flow Cytometry to analyse tissue infiltrating immune cells and cytokine production during psoriasis using mouse models.
Wed Jun 23 00:00:00 UTC 2021
Mucosal-associated invariant T cells promote inflammation and intestinal dysbiosis leading to metabolic dysfunction during obesity.
Nature communications
Toubal A,Kiaf B,Beaudoin L,Cagninacci L,Rhimi M,Fruchet B,da Silva J,Corbett AJ,Simoni Y,Lantz O,Rossjohn J,McCluskey J,Lesnik P,Maguin E,Lehuen A
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jul 24 00:00:00 UTC 2020
CD71+ Erythroid Cells in Human Neonates Exhibit Immunosuppressive Properties and Compromise Immune Response Against Systemic Infection in Neonatal Mice.
Frontiers in immunology
Elahi S,Vega-López MA,Herman-Miguel V,Ramírez-Estudillo C,Mancilla-Ramírez J,Motyka B,West L,Oyegbami O
13-0114-82 was used in Flow Cytometry to study the frequency and functionality of CD71+ erythroid cells in the peripheral blood of human newborns/children from 1-day to 5-year and compared them with their counterparts in the neonatal mice.
Mon Dec 28 00:00:00 UTC 2020
An Nfil3-Zeb2-Id2 pathway imposes Irf8 enhancer switching during cDC1 development.
Nature immunology
Bagadia P,Huang X,Liu TT,Durai V,Grajales-Reyes GE,Nitschké M,Modrusan Z,Granja JM,Satpathy AT,Briseño CG,Gargaro M,Iwata A,Kim S,Chang HY,Shaw AS,Murphy TL,Murphy KM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Sep 01 00:00:00 UTC 2019
Peroxisome Proliferator-activated Receptor-γ Deficiency Exacerbates Fibrotic Response to Mycobacteria Peptide in Murine Sarcoidosis Model.
American journal of respiratory cell and molecular biology
Malur A,Mohan A,Barrington RA,Leffler N,Malur A,Muller-Borer B,Murray G,Kew K,Zhou C,Russell J,Jones JL,Wingard CJ,Barna BP,Thomassen MJ
13-0114-82 was used in Immunohistochemistry to investigate if the addition of mycobacterial ESAT-6 (early secreted antigenic target protein 6) to multiwall carbon nanotube (MWCNT)-elicited chronic granulomatous disease might exacerbate pulmonary granulomatous pathology.
Thu Aug 01 00:00:00 UTC 2019
CD1d highly expressed on DCs reduces lung tumor burden by enhancing antitumor immunity.
Oncology reports
Li Y,Zhao C,Liu J,Lu Z,Lu M,Gu J,Liu R
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed May 01 00:00:00 UTC 2019
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Oct 14 00:00:00 UTC 2020
Knockdown of Ant2 Reduces Adipocyte Hypoxia And Improves Insulin Resistance in Obesity.
Nature metabolism
Seo JB,Riopel M,Cabrales P,Huh JY,Bandyopadhyay GK,Andreyev AY,Murphy AN,Beeman SC,Smith GI,Klein S,Lee YS,Olefsky JM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2019
Integrated Cross-Species Analysis Identifies a Conserved Transitional Dendritic Cell Population.
Cell reports
Leylek R,Alcántara-Hernández M,Lanzar Z,Lüdtke A,Perez OA,Reizis B,Idoyaga J
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Dec 10 00:00:00 UTC 2019
Nidogen-1 Contributes to the Interaction Network Involved in Pro-B Cell Retention in the Peri-sinusoidal Hematopoietic Stem Cell Niche.
Cell reports
Balzano M,De Grandis M,Vu Manh TP,Chasson L,Bardin F,Farina A,Sergé A,Bidaut G,Charbord P,Hérault L,Bailly AL,Cartier-Michaud A,Boned A,Dalod M,Duprez E,Genever P,Coles M,Bajenoff M,Xerri L,Aurrand-Lions M,Schiff C,Mancini SJC
13-0114 was used in Flow cytometry/Cell sorting to study pro-B cells in their support of stroll cells and their interaction network allowing their retention.
Tue Mar 19 00:00:00 UTC 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jan 25 00:00:00 UTC 2018
NK Cells Stimulate Recruitment of cDC1 into the Tumor Microenvironment Promoting Cancer Immune Control.
Cell
Böttcher JP,Bonavita E,Chakravarty P,Blees H,Cabeza-Cabrerizo M,Sammicheli S,Rogers NC,Sahai E,Zelenay S,Reis e Sousa C
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Feb 22 00:00:00 UTC 2018
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jun 28 00:00:00 UTC 2018
Interleukin-17-Producing γδ T Cells Originate from SOX13+ Progenitors that Are Independent of γδTCR Signaling.
Immunity
Spidale NA,Sylvia K,Narayan K,Miu B,Frascoli M,Melichar HJ,Zhihao W,Kisielow J,Palin A,Serwold T,Love P,Kobayashi M,Yoshimoto M,Jain N,Kang J
13-0114-82 was used in Flow Cytometry to generate reporter mice for the Tγδ17 lineage-defining transcription factor SOX13 and identify fetal-origin, intrathymic Sox13+ progenitors.
Tue Nov 20 00:00:00 UTC 2018
Insulin resistance causes inflammation in adipose tissue.
The Journal of clinical investigation
Shimobayashi M,Albert V,Woelnerhanssen B,Frei IC,Weissenberger D,Meyer-Gerspach AC,Clement N,Moes S,Colombi M,Meier JA,Swierczynska MM,Jenö P,Beglinger C,Peterli R,Hall MN
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Apr 02 00:00:00 UTC 2018
Granulocyte-CSF links destructive inflammation and comorbidities in obstructive lung disease.
The Journal of clinical investigation
Tsantikos E,Lau M,Castelino CM,Maxwell MJ,Passey SL,Hansen MJ,McGregor NE,Sims NA,Steinfort DP,Irving LB,Anderson GP,Hibbs ML
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jun 01 00:00:00 UTC 2018
Photodynamic therapy reduces the inhibitory effect of osteosarcoma cells on dendritic cells by upregulating HSP70.
Oncology letters
Zhang F,Zhu Y,Fan G,Hu S
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2018
FACS isolation of live mouse eosinophils at high purity via a protocol that does not target Siglec F.
Journal of immunological methods
Geslewitz WE,Percopo CM,Rosenberg HF
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2018
Macrophage Depletion Protects against Cigarette Smoke-Induced Inflammatory Response in the Mouse Colon and Lung.
Frontiers in physiology
Lim D,Kim W,Lee C,Bae H,Kim J
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Characterization of Multiple Cytokine Combinations and TGF-β on Differentiation and Functions of Myeloid-Derived Suppressor Cells.
International journal of molecular sciences
Lee CR,Lee W,Cho SK,Park SG
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Mar 15 00:00:00 UTC 2018
Lung Epithelial Cells Coordinate Innate Lymphocytes and Immunity against Pulmonary Fungal Infection.
Cell host & microbe
Hernández-Santos N,Wiesner DL,Fites JS,McDermott AJ,Warner T,Wüthrich M,Klein BS
13-0114-82 was used in Flow Cytometry to demonstrate that NF-κB signaling in LECs is essential for immunity against the pulmonary fungal pathogen Blastomyces dermatitidis.
Wed Apr 11 00:00:00 UTC 2018
A stochastic epigenetic switch controls the dynamics of T-cell lineage commitment.
eLife
Ng KK,Yui MA,Mehta A,Siu S,Irwin B,Pease S,Hirose S,Elowitz MB,Rothenberg EV,Kueh HY
13-0114 was used in Flow cytometry/Cell sorting to study the activation of Bcl11b, which controls T-cell fate commitment.
Tue Nov 20 00:00:00 UTC 2018
Epigenetic Restoration of Fetal-like IGF1 Signaling Inhibits Leukemia Stem Cell Activity.
Cell stem cell
Giambra V,Gusscott S,Gracias D,Song R,Lam SH,Panelli P,Tyshchenko K,Jenkins CE,Hoofd C,Lorzadeh A,Carles A,Hirst M,Eaves CJ,Weng AP
13-0114-82 was used in Flow Cytometry to demonstrate that resurrecting dormant fetal programs in adult cells may represent an alternate therapeutic approach in human cancer.
Thu Nov 01 00:00:00 UTC 2018
Notch Signaling Facilitates In Vitro Generation of Cross-Presenting Classical Dendritic Cells.
Cell reports
Kirkling ME,Cytlak U,Lau CM,Lewis KL,Resteu A,Khodadadi-Jamayran A,Siebel CW,Salmon H,Merad M,Tsirigos A,Collin M,Bigley V,Reizis B
Tue Jun 19 00:00:00 UTC 2018
Combined antiangiogenic and anti-PD-L1 therapy stimulates tumor immunity through HEV formation.
Science translational medicine
Allen E,Jabouille A,Rivera LB,Lodewijckx I,Missiaen R,Steri V,Feyen K,Tawney J,Hanahan D,Michael IP,Bergers G
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Wed Apr 12 00:00:00 UTC 2017
RIG-I antiviral signaling drives interleukin-23 production and psoriasis-like skin disease.
EMBO molecular medicine
Zhu H,Lou F,Yin Q,Gao Y,Sun Y,Bai J,Xu Z,Liu Z,Cai W,Ke F,Zhang L,Zhou H,Wang H,Wang G,Chen X,Zhang H,Wang Z,Ginhoux F,Lu C,Su B,Wang H
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017
FcεRI γ-Chain Negatively Modulates Dectin-1 Responses in Dendritic Cells.
Frontiers in immunology
Pan YG,Yu YL,Lin CC,Lanier LL,Chu CL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Nov 20 00:00:00 UTC 2019
Cornea lymphatics drive the CD8+ T-cell response to herpes simplex virus-1.
Immunology and cell biology
Gurung HR,Carr MM,Carr DJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Jan 01 00:00:00 UTC 2017
An Anti-Inflammatory Role for NLRP10 in Murine Cutaneous Leishmaniasis.
Journal of immunology (Baltimore, Md. : 1950)
Clay GM,Valadares DG,Graff JW,Ulland TK,Davis RE,Scorza BM,Zhanbolat BS,Chen Y,Sutterwala FS,Wilson ME
13-0114 was used in Flow cytometry/Cell sorting to determine the role of NLRP10 cutaneous leishmaniasis.
Sun Oct 15 00:00:00 UTC 2017
B Cell-Extrinsic Myd88 and Fcer1g Negatively Regulate Autoreactive and Normal B Cell Immune Responses.
Journal of immunology (Baltimore, Md. : 1950)
Sweet RA,Nickerson KM,Cullen JL,Wang Y,Shlomchik MJ
Tue Aug 01 00:00:00 UTC 2017
Identification of different macrophage subpopulations with distinct activities in a mouse model of oxygen-induced retinopathy.
International journal of molecular medicine
Zhu Y,Zhang L,Lu Q,Gao Y,Cai Y,Sui A,Su T,Shen X,Xie B
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Tue Aug 01 00:00:00 UTC 2017
Chronic Plasmodium chabaudi Infection Generates CD4 Memory T Cells with Increased T Cell Receptor Sensitivity but Poor Secondary Expansion and Increased Apoptosis.
Infection and immunity
Opata MM,Stephens R
Wed Mar 01 00:00:00 UTC 2017
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2017
The Second-Generation PGI2 Analogue Treprostinil Fails to Chemoprevent Tumors in a Murine Lung Adenocarcinoma Model.
Cancer prevention research (Philadelphia, Pa.)
Dwyer-Nield L,Hickey GA,Friedman M,Choo K,McArthur DG,Tennis MA,New ML,Geraci M,Keith RL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2017
Calreticulin is an effective immunologic adjuvant to tumor-associated antigens.
Experimental and therapeutic medicine
Wang J,Gao ZP,Qin S,Liu CB,Zou LL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2017
PEDF mediates pathological neovascularization by regulating macrophage recruitment and polarization in the mouse model of oxygen-induced retinopathy.
Scientific reports
Gao S,Li C,Zhu Y,Wang Y,Sui A,Zhong Y,Xie B,Shen X
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Feb 17 00:00:00 UTC 2017
PEDF mediates pathological neovascularization by regulating macrophage recruitment and polarization in the mouse model of oxygen-induced retinopathy.
Scientific reports
Gao S,Li C,Zhu Y,Wang Y,Sui A,Zhong Y,Xie B,Shen X
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Feb 17 00:00:00 UTC 2017
CD11c+ resident macrophages drive hepatocyte death-triggered liver fibrosis in a murine model of nonalcoholic steatohepatitis.
JCI insight
Itoh M,Suganami T,Kato H,Kanai S,Shirakawa I,Sakai T,Goto T,Asakawa M,Hidaka I,Sakugawa H,Ohnishi K,Komohara Y,Asano K,Sakaida I,Tanaka M,Ogawa Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Thu Nov 16 00:00:00 UTC 2017
Precision-cut Mouse Lung Slices to Visualize Live Pulmonary Dendritic Cells.
Journal of visualized experiments : JoVE
Lyons-Cohen MR,Thomas SY,Cook DN,Nakano H
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Wed Apr 05 00:00:00 UTC 2017
BEX1 is an RNA-dependent mediator of cardiomyopathy.
Nature communications
Accornero F,Schips TG,Petrosino JM,Gu SQ,Kanisicak O,van Berlo JH,Molkentin JD
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Nov 30 00:00:00 UTC 2017
Commensal microbiota maintains alveolar macrophages with a low level of CCL24 production to generate anti-metastatic tumor activity.
Scientific reports
Cheng M,Chen Y,Wang L,Chen W,Yang L,Shen G,Xu T,Shen G,Tian Z,Hu S
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Aug 07 00:00:00 UTC 2017
Batf3-dependent CD8α+ Dendritic Cells Aggravates Atherosclerosis via Th1 Cell Induction and Enhanced CCL5 Expression in Plaque Macrophages.
EBioMedicine
Li Y,Liu X,Duan W,Tian H,Zhu G,He H,Yao S,Yi S,Song W,Tang H
13-0114 was used in Immunohistochemistry to show that Batf3-/-Apoe-/- mice, lacking CD8α+ DCs, exhibited a significant reduction in atherogenesis and T help 1 (Th1) cells compared with Apoe-/- controls.
Sat Apr 01 00:00:00 UTC 2017
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Oct 06 00:00:00 UTC 2016
Acidic chitinase primes the protective immune response to gastrointestinal nematodes.
Nature immunology
Vannella KM,Ramalingam TR,Hart KM,de Queiroz Prado R,Sciurba J,Barron L,Borthwick LA,Smith AD,Mentink-Kane M,White S,Thompson RW,Cheever AW,Bock K,Moore I,Fitz LJ,Urban JF,Wynn TA
13-0114 was used in Flow cytometry/Cell sorting to use AMCase-deficient mice as a means to dissect the role of the enzyme in several models of helminth infection and type 2 cytokine-driven airway inflammation.
Sun May 01 00:00:00 UTC 2016
Loss of gastrokine-2 drives premalignant gastric inflammation and tumor progression.
The Journal of clinical investigation
Menheniott TR,O'Connor L,Chionh YT,Däbritz J,Scurr M,Rollo BN,Ng GZ,Jacobs S,Catubig A,Kurklu B,Mercer S,Minamoto T,Ong DE,Ferrero RL,Fox JG,Wang TC,Sutton P,Judd LM,Giraud AS
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Apr 01 00:00:00 UTC 2016
IL-23 induced in keratinocytes by endogenous TLR4 ligands polarizes dendritic cells to drive IL-22 responses to skin immunization.
The Journal of experimental medicine
Yoon J,Leyva-Castillo JM,Wang G,Galand C,Oyoshi MK,Kumar L,Hoff S,He R,Chervonsky A,Oppenheim JJ,Kuchroo VK,van den Brink MR,Malefyt Rde W,Tessier PA,Fuhlbrigge R,Rosenstiel P,Terhorst C,Murphy G,Geha RS
13-0114 was used in Flow cytometry/Cell sorting to elucidate the mechanisms behind DC-derived IL-23 in a model of skin inflammation.
Mon Sep 19 00:00:00 UTC 2016
Dendritic cell sphingosine-1-phosphate lyase regulates thymic egress.
The Journal of experimental medicine
Zamora-Pineda J,Kumar A,Suh JH,Zhang M,Saba JD
13-0114 was used in Immunofluorescence to investigate the role of dendritic cell sphingosine-1-phosphate lyase in thymic egress.
Mon Nov 14 00:00:00 UTC 2016
Macrophages transfer antigens to dendritic cells by releasing exosomes containing dead-cell-associated antigens partially through a ceramide-dependent pathway to enhance CD4(+) T-cell responses.
Immunology
Xu Y,Liu Y,Yang C,Kang L,Wang M,Hu J,He H,Song W,Tang H
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sat Oct 01 00:00:00 UTC 2016
Identification of a Hematopoietic Cell Dedifferentiation-Inducing Factor.
Journal of cellular physiology
Li Y,Adomat H,Guns ET,Hojabrpour P,Duronio V,Curran TA,Jalili RB,Jia W,Delwar Z,Zhang Y,Elizei SS,Ghahary A
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunocytochemistry
Wed Jun 01 00:00:00 UTC 2016
Age-dependent divergent effects of OX40L treatment on the development of diabetes in NOD mice.
Autoimmunity
Haddad CS,Bhattacharya P,Alharshawi K,Marinelarena A,Kumar P,El-Sayed O,Elshabrawy HA,Epstein AL,Prabhakar BS
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
CD80 and CD86 knockdown in dendritic cells regulates Th1/Th2 cytokine production in asthmatic mice.
Experimental and therapeutic medicine
Li JG,DU YM,Yan ZD,Yan J,Zhuansun YX,Chen R,Zhang W,Feng SL,Ran PX
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Flagellin is a strong vaginal adjuvant of a therapeutic vaccine for genital cancer.
Oncoimmunology
Lee SE,Hong SH,Verma V,Lee YS,Duong TN,Jeong K,Uthaman S,Sung YC,Lee JT,Park IK,Min JJ,Rhee JH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Mon Feb 01 00:00:00 UTC 2016
Flagellin is a strong vaginal adjuvant of a therapeutic vaccine for genital cancer.
Oncoimmunology
Lee SE,Hong SH,Verma V,Lee YS,Duong TN,Jeong K,Uthaman S,Sung YC,Lee JT,Park IK,Min JJ,Rhee JH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Mon Feb 01 00:00:00 UTC 2016
Antigen presenting cell abnormalities in the Cln3(-/-) mouse model of juvenile neuronal ceroid lipofuscinosis.
Biochimica et biophysica acta
Hersrud SL,Kovács AD,Pearce DA
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunocytochemistry
Fri Jul 01 00:00:00 UTC 2016
Epigallocatechin-3-gallate (EGCG) inhibits imiquimod-induced psoriasis-like inflammation of BALB/c mice.
BMC complementary and alternative medicine
Zhang S,Liu X,Mei L,Wang H,Fang F
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Aug 31 00:00:00 UTC 2016
Induction of Suppressor Cells and Increased Tumor Growth following Chronic Psychosocial Stress in Male Mice.
PloS one
Schmidt D,Peterlik D,Reber SO,Lechner A,Männel DN
13-0114 was used in Flow cytometry/Cell sorting to observe the immunological effects of chronic psychosocial stress in mice.
Fri Aug 18 00:00:00 UTC 2017
Altered gp130 signalling ameliorates experimental colitis via myeloid cell-specific STAT3 activation and myeloid-derived suppressor cells.
Scientific reports
Däbritz J,Judd LM,Chalinor HV,Menheniott TR,Giraud AS
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Feb 05 00:00:00 UTC 2016
Thymic exosomes promote the final maturation of thymocytes.
Scientific reports
Lundberg V,Berglund M,Skogberg G,Lindgren S,Lundqvist C,Gudmundsdottir J,Thörn K,Telemo E,Ekwall O
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Nov 08 00:00:00 UTC 2016
Epidermal RAF prevents allergic skin disease.
eLife
Raguz J,Jeric I,Niault T,Nowacka JD,Kuzet SE,Rupp C,Fischer I,Biggi S,Borsello T,Baccarini M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Jul 19 00:00:00 UTC 2016
Diet-dependent, microbiota-independent regulation of IL-10-producing lamina propria macrophages in the small intestine.
Scientific reports
Ochi T,Feng Y,Kitamoto S,Nagao-Kitamoto H,Kuffa P,Atarashi K,Honda K,Teitelbaum DH,Kamada N
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Jun 15 00:00:00 UTC 2016
KLF4-dependent phenotypic modulation of smooth muscle cells has a key role in atherosclerotic plaque pathogenesis.
Nature medicine
Shankman LS,Gomez D,Cherepanova OA,Salmon M,Alencar GF,Haskins RM,Swiatlowska P,Newman AA,Greene ES,Straub AC,Isakson B,Randolph GJ,Owens GK
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Unique role for ATG5 in neutrophil-mediated immunopathology during M. tuberculosis infection.
Nature
Kimmey JM,Huynh JP,Weiss LA,Park S,Kambal A,Debnath J,Virgin HW,Stallings CL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Dec 24 00:00:00 UTC 2015
MET is required for the recruitment of anti-tumoural neutrophils.
Nature
Finisguerra V,Di Conza G,Di Matteo M,Serneels J,Costa S,Thompson AA,Wauters E,Walmsley S,Prenen H,Granot Z,Casazza A,Mazzone M
13-0114 was used in Immunohistochemistry on paraffin embedded tissues to identify a role for the MET proto-oncogene in anti-tumoural neutrophil recruitment.
Thu Jun 18 00:00:00 UTC 2015
MET is required for the recruitment of anti-tumoural neutrophils.
Nature
Finisguerra V,Di Conza G,Di Matteo M,Serneels J,Costa S,Thompson AA,Wauters E,Walmsley S,Prenen H,Granot Z,Casazza A,Mazzone M
13-0114 was used in Immunohistochemistry on paraffin embedded tissues to identify a role for the MET proto-oncogene in anti-tumoural neutrophil recruitment.
Thu Jun 18 00:00:00 UTC 2015
Injectable, spontaneously assembling, inorganic scaffolds modulate immune cells in vivo and increase vaccine efficacy.
Nature biotechnology
Kim J,Li WA,Choi Y,Lewin SA,Verbeke CS,Dranoff G,Mooney DJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Imaging Macrophage and Hematopoietic Progenitor Proliferation in Atherosclerosis.
Circulation research
Ye YX,Calcagno C,Binderup T,Courties G,Keliher EJ,Wojtkiewicz GR,Iwamoto Y,Tang J,Pérez-Medina C,Mani V,Ishino S,Johnbeck CB,Knigge U,Fayad ZA,Libby P,Weissleder R,Tawakol A,Dubey S,Belanger AP,Di Carli MF,Swirski FK,Kjaer A,Mulder WJ,Nahrendorf M
13-0114 was used in Flow cytometry/Cell sorting to perform (18)F-FLT positron emission tomography-computed tomography was performed in mice, rabbits, and humans with atherosclerosis.
Fri Oct 23 00:00:00 UTC 2015
Distinct germinal center selection at local sites shapes memory B cell response to viral escape.
The Journal of experimental medicine
Adachi Y,Onodera T,Yamada Y,Daio R,Tsuiji M,Inoue T,Kobayashi K,Kurosaki T,Ato M,Takahashi Y
13-0114 was used in Flow cytometry/Cell sorting to investigate the cellular events dictating the cross-reactive memory B cell responses, showing that distinct germinal center selection at local sites shapes response to viral escape.
Mon Sep 21 00:00:00 UTC 2015
BAFF activation of the ERK5 MAP kinase pathway regulates B cell survival.
The Journal of experimental medicine
Jacque E,Schweighoffer E,Tybulewicz VL,Ley SC
13-0114 was used in Flow cytometry/Cell sorting to reveal a potentially critical role for the MEK5-ERK5 MAP kinase signalling pathway in BAFF-induced mature B cell survival and homeostatic maintenance of B2 cell numbers.
Mon Jun 01 00:00:00 UTC 2015
Phospholipid oxidation generates potent anti-inflammatory lipid mediators that mimic structurally related pro-resolving eicosanoids by activating Nrf2.
EMBO molecular medicine
Bretscher P,Egger J,Shamshiev A,Trötzmüller M,Köfeler H,Carreira EM,Kopf M,Freigang S
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri May 01 00:00:00 UTC 2015
Atypical chemokine receptor 1 deficiency reduces atherogenesis in ApoE-knockout mice.
Cardiovascular research
Wan W,Liu Q,Lionakis MS,Marino AP,Anderson SA,Swamydas M,Murphy PM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Single-Dose CpG Immunization Protects Against a Heterosubtypic Challenge and Generates Antigen-Specific Memory T Cells.
Frontiers in immunology
Vogel AJ,Brown DM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jul 10 00:00:00 UTC 2015
LynA regulates an inflammation-sensitive signaling checkpoint in macrophages.
eLife
Freedman TS,Tan YX,Skrzypczynska KM,Manz BN,Sjaastad FV,Goodridge HS,Lowell CA,Weiss A
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Oct 30 00:00:00 UTC 2015
The late endosomal adaptor molecule p14 (LAMTOR2) regulates TGFβ1-mediated homeostasis of Langerhans cells.
The Journal of investigative dermatology
Sparber F,Tripp CH,Komenda K,Scheffler JM,Clausen BE,Huber LA,Romani N,Stoitzner P
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Intermuscular and perimuscular fat expansion in obesity correlates with skeletal muscle T cell and macrophage infiltration and insulin resistance.
International journal of obesity (2005)
Khan IM,Perrard XY,Brunner G,Lui H,Sparks LM,Smith SR,Wang X,Shi ZZ,Lewis DE,Wu H,Ballantyne CM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2015
A novel regulatory macrophage induced by a helminth molecule instructs IL-10 in CD4+ T cells and protects against mucosal inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Ziegler T,Rausch S,Steinfelder S,Klotz C,Hepworth MR,Kühl AA,Burda PC,Lucius R,Hartmann S
13-0114 was used in Magnetic cell separation to investigate the mechanism by which the helminth immunomodulator AvCystatin induces a novel regulatory macrophage able to protect against allergic mucosal inflammation in mice.
Sun Feb 15 00:00:00 UTC 2015
DGCR14 induces Il17a gene expression through the RORγ/BAZ1B/RSKS2 complex.
Molecular and cellular biology
Takada I
13-0114 was used in Magnetic cell separation to show the roles of DGCR14, RSK2, and BAZ1B in the transcriptional regulation of Il17a mRNA during TH17 cell differentiation.
Thu Jan 01 00:00:00 UTC 2015
A far-red fluorescent probe for flow cytometry and image-based functional studies of xenobiotic sequestering macrophages.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Keswani RK,Yoon GS,Sud S,Stringer KA,Rosania GR
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Adjuvants may reduce in vivo transfection levels for DNA vaccination in mice leading to reduced antigen-specific CD8+ T cell responses.
Human vaccines & immunotherapeutics
Rose AH,Hoffmann FW,Hara JH,Urschitz J,Moisyadi S,Hoffmann PR,Bertino P
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon May 16 00:00:00 UTC 2016
Inadequate activation of the HBsAg-specific Th cells by APCs leads to hyporesponsiveness to HBsAg vaccine in B10.S mice.
Human vaccines & immunotherapeutics
Li X,Xu J,Lv Z,Wang J,Sun S,Zhu W,Wang B,He R,Qu D
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Apr 19 00:00:00 UTC 2016
Anti-tumor effect of the alphavirus-based virus-like particle vector expressing prostate-specific antigen in a HLA-DR transgenic mouse model of prostate cancer.
Vaccine
Riabov V,Tretyakova I,Alexander RB,Pushko P,Klyushnenkova EN
13-0114 was used in Immunofluorescence to determine whether an alphavirus-based vaccine encoding human Prostate-Specific Antigen could generate an effective anti-tumour immune response in a mouse model of prostate cancer.
Mon Oct 05 00:00:00 UTC 2015
Reducing progression of experimental lupus nephritis via inhibition of the B7/CD28 signaling pathway.
Molecular medicine reports
Huang L,Kong Y,Wang J,Sun J,Shi Q,Qiu YH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Advanced Glycation End Products Enhance Macrophages Polarization into M1 Phenotype through Activating RAGE/NF-κB Pathway.
BioMed research international
Jin X,Yao T,Zhou Z,Zhu J,Zhang S,Hu W,Shen C
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Mar 18 00:00:00 UTC 2016
Targeted Mutation of Nuclear Bone Morphogenetic Protein 2 Impairs Secondary Immune Response in a Mouse Model.
BioMed research international
Olsen DS,Goar WA,Nichols BA,Bailey KT,Christensen SL,Merriam KR,Reynolds PR,Wilson E,Weber KS,Bridgewater LC
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
M cell-derived vesicles suggest a unique pathway for trans-epithelial antigen delivery.
Tissue barriers
Sakhon OS,Ross B,Gusti V,Pham AJ,Vu K,Lo DD
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Fri Apr 03 00:00:00 UTC 2015
M cell-derived vesicles suggest a unique pathway for trans-epithelial antigen delivery.
Tissue barriers
Sakhon OS,Ross B,Gusti V,Pham AJ,Vu K,Lo DD
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Fri Apr 03 00:00:00 UTC 2015
Granulocyte/Macrophage Colony-stimulating Factor-dependent Dendritic Cells Restrain Lean Adipose Tissue Expansion.
The Journal of biological chemistry
Pamir N,Liu NC,Irwin A,Becker L,Peng Y,Ronsein GE,Bornfeldt KE,Duffield JS,Heinecke JW
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jun 05 00:00:00 UTC 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Sep 15 00:00:00 UTC 2015
Disruption of p21-activated kinase 1 gene diminishes atherosclerosis in apolipoprotein E-deficient mice.
Nature communications
Singh NK,Kotla S,Dyukova E,Traylor JG,Orr AW,Chernoff J,Marion TN,Rao GN
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Wed Jun 24 00:00:00 UTC 2015
Eosinophils from Murine Lamina Propria Induce Differentiation of Naïve T Cells into Regulatory T Cells via TGF-β1 and Retinoic Acid.
PloS one
Chen HH,Sun AH,Ojcius DM,Hu WL,Ge YM,Lin X,Li LJ,Pan JP,Yan J
13-0114 was used in Flow cytometry/Cell sorting to investigate mechanisms that induce Tregs, showing that eosinophils from murine lamina propria induce differentiation via TGF-β1 and retinoic acid.
Fri Jun 17 00:00:00 UTC 2016
IFN-γ and IL-21 Double Producing T Cells Are Bcl6-Independent and Survive into the Memory Phase in Plasmodium chabaudi Infection.
PloS one
Carpio VH,Opata MM,Montañez ME,Banerjee PP,Dent AL,Stephens R
13-0114 was used in Flow cytometry/Cell sorting to elucidate the lineage of a specific Ifng+ CD4 T cell subset generated during Plasmodium chabaudi infection in mice.
Sun Jun 26 00:00:00 UTC 2016
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Mon Dec 01 00:00:00 UTC 2014
Immune complexes stimulate CCR7-dependent dendritic cell migration to lymph nodes.
Nature medicine
Clatworthy MR,Aronin CE,Mathews RJ,Morgan NY,Smith KG,Germain RN
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Mon Dec 01 00:00:00 UTC 2014
Sessile alveolar macrophages communicate with alveolar epithelium to modulate immunity.
Nature
Westphalen K,Gusarova GA,Islam MN,Subramanian M,Cohen TS,Prince AS,Bhattacharya J
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Thu Feb 27 00:00:00 UTC 2014
High-fat-diet-mediated dysbiosis promotes intestinal carcinogenesis independently of obesity.
Nature
Schulz MD,Atay C,Heringer J,Romrig FK,Schwitalla S,Aydin B,Ziegler PK,Varga J,Reindl W,Pommerenke C,Salinas-Riester G,Böck A,Alpert C,Blaut M,Polson SC,Brandl L,Kirchner T,Greten FR,Polson SW,Arkan MC
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Oct 23 00:00:00 UTC 2014
CLEC-2 is required for development and maintenance of lymph nodes.
Blood
Bénézech C,Nayar S,Finney BA,Withers DR,Lowe K,Desanti GE,Marriott CL,Watson SP,Caamaño JH,Buckley CD,Barone F
13-0114 was used in Immunofluorescence to investigate the role that CLEC-2 plays in the development and maintenance of lymph nodes.
Thu May 15 00:00:00 UTC 2014
Imaging murine NALT following intranasal immunization with flagellin-modified circumsporozoite protein malaria vaccines.
Mucosal immunology
Nacer A,Carapau D,Mitchell R,Meltzer A,Shaw A,Frevert U,Nardin EH
13-0114 was used in Flow cytometry/Cell sorting to understand the intranasal immunisation-elicited immune response in nasopharynx-associated lymphoid tissue.
Sat Mar 01 00:00:00 UTC 2014
Specific microbiota-induced intestinal Th17 differentiation requires MHC class II but not GALT and mesenteric lymph nodes.
Journal of immunology (Baltimore, Md. : 1950)
Geem D,Medina-Contreras O,McBride M,Newberry RD,Koni PA,Denning TL
13-0114 was used in Magnetic cell separation to investigate whether GALT and/or mesenteric lymph nodes are required for intestinal Th17 differentiation, showing that specific microbiota-induced intestinal Th17 differentiation requires MHC class II.
Tue Jul 01 00:00:00 UTC 2014
Relapsing-remitting central nervous system autoimmunity mediated by GFAP-specific CD8 T cells.
Journal of immunology (Baltimore, Md. : 1950)
Sasaki K,Bean A,Shah S,Schutten E,Huseby PG,Peters B,Shen ZT,Vanguri V,Liggitt D,Huseby ES
13-0114 was used in Immunohistochemistry to investigate the mechanism by which CD8 T cells mediate relapsing-remitting inflammatory CNS autoimmunity.
Tue Apr 01 00:00:00 UTC 2014
Mice deficient in intestinal epithelium cytochrome P450 reductase are prone to acute toxin-induced mucosal damage.
Scientific reports
Ahlawat S,Xie F,Zhu Y,D'Hondt R,Ding X,Zhang QY,Mantis NJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Thu Jul 03 00:00:00 UTC 2014
A chitin-like component on sclerotic cells of Fonsecaea pedrosoi inhibits Dectin-1-mediated murine Th17 development by masking β-glucans.
PloS one
Dong B,Li D,Li R,Chen SC,Liu W,Liu W,Chen L,Chen Y,Zhang X,Tong Z,Xia Y,Xia P,Wang Y,Duan Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2015
P2X7 receptor modulates inflammatory and functional pulmonary changes induced by silica.
PloS one
Monção-Ribeiro LC,Faffe DS,Santana PT,Vieira FS,da Graça CL,Marques-da-Silva C,Machado MN,Caruso-Neves C,Zin WA,Borojevic R,Takiya CM,Coutinho-Silva R
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Sep 17 00:00:00 UTC 2015
A type I IFN-Flt3 ligand axis augments plasmacytoid dendritic cell development from common lymphoid progenitors.
The Journal of experimental medicine
Chen YL,Chen TT,Pai LM,Wesoly J,Bluyssen HA,Lee CK
13-0114 was used in Flow cytometry/Cell sorting to define a critical role for the FL/IFN-I/Flt3 axis in plasmacytoid dendritic cell differentiation from common lymphoid progenitors.
Mon Nov 18 00:00:00 UTC 2013
Genetic deletion of chemokine receptor Ccr7 exacerbates atherogenesis in ApoE-deficient mice.
Cardiovascular research
Wan W,Lionakis MS,Liu Q,Roffê E,Murphy PM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2013
CX(3)CR1(+) macrophages support IL-22 production by innate lymphoid cells during infection with Citrobacter rodentium.
Mucosal immunology
Manta C,Heupel E,Radulovic K,Rossini V,Garbi N,Riedel CU,Niess JH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2013
CX(3)CR1(+) macrophages support IL-22 production by innate lymphoid cells during infection with Citrobacter rodentium.
Mucosal immunology
Manta C,Heupel E,Radulovic K,Rossini V,Garbi N,Riedel CU,Niess JH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2013
Regulatory T cells accumulate and proliferate in the ischemic hemisphere for up to 30 days after MCAO.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Stubbe T,Ebner F,Richter D,Engel O,Klehmet J,Royl G,Meisel A,Nitsch R,Meisel C,Brandt C
13-0114 was used in Immunohistochemistry to study immune cell distribution and function after middle cerebral artery occlusion.
Tue Jan 01 00:00:00 UTC 2013
Hepatic crown-like structure: a unique histological feature in non-alcoholic steatohepatitis in mice and humans.
PloS one
Itoh M,Kato H,Suganami T,Konuma K,Marumoto Y,Terai S,Sakugawa H,Kanai S,Hamaguchi M,Fukaishi T,Aoe S,Akiyoshi K,Komohara Y,Takeya M,Sakaida I,Ogawa Y
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Sun Oct 12 00:00:00 UTC 2014
Thymic stromal lymphopoietin attenuates the development of atherosclerosis in ApoE-/- mice.
Journal of the American Heart Association
Yu K,Zhu P,Dong Q,Zhong Y,Zhu Z,Lin Y,Huang Y,Meng K,Ji Q,Yi G,Zhang W,Wu B,Mao Y,Cheng P,Zhao X,Mao X,Zeng Q
13-0114 was used in Flow cytometry/Cell sorting to examine the phenotype of TSLP-conditioned bone marrow dendritic cells of apolipoprotein E-deficient mice and their capacity to induce the differentiation of Tregs.
Wed Aug 28 00:00:00 UTC 2013
Emulsified phosphatidylserine, simple and effective peptide carrier for induction of potent epitope-specific T cell responses.
PloS one
Ichihashi T,Satoh T,Sugimoto C,Kajino K
13-0114 was used in Flow cytometry/Cell sorting to show that phosphatidylserine is a promising novel adjuvant for T cell-inducing peptide vaccines.
Wed Sep 11 00:00:00 UTC 2013
Neutralization of TSLP inhibits airway remodeling in a murine model of allergic asthma induced by chronic exposure to house dust mite.
PloS one
Chen ZG,Zhang TT,Li HT,Chen FH,Zou XL,Ji JZ,Chen H
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Aug 20 00:00:00 UTC 2013
Inflammatory arthritis increases mouse osteoclast precursors with myeloid suppressor function.
The Journal of clinical investigation
Charles JF,Hsu LY,Niemi EC,Weiss A,Aliprantis AO,Nakamura MC
13-0114 was used in Flow cytometry/Cell sorting to identify a CD11b(-/lo)Ly6C(hi) bone marrow population with osteoclast precursor activity.
Sat Dec 01 00:00:00 UTC 2012
Notch, Id2, and RORγt sequentially orchestrate the fetal development of lymphoid tissue inducer cells.
The Journal of experimental medicine
Cherrier M,Sawa S,Eberl G
13-0114 was used in Magnetic cell separation to investigate the development of lymphoid tissue inducer (LTi) cells from common lymphoid progenitors (CLPs) in the fetal liver, showing that sequential changes in expression of Notch, Id2, and RORγt orchestrate the development of LTi cells.
Mon Apr 09 00:00:00 UTC 2012
Alum increases antigen uptake, reduces antigen degradation and sustains antigen presentation by DCs in vitro.
Immunology letters
Ghimire TR,Benson RA,Garside P,Brewer JM
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2012
Differential gene expression in mice infected with distinct Toxoplasma strains.
Infection and immunity
Hill RD,Gouffon JS,Saxton AM,Su C
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2012
Depletion of B2 but not B1a B cells in BAFF receptor-deficient ApoE mice attenuates atherosclerosis by potently ameliorating arterial inflammation.
PloS one
Kyaw T,Tay C,Hosseini H,Kanellakis P,Gadowski T,MacKay F,Tipping P,Bobik A,Toh BH
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunohistochemistry
Mon May 14 00:00:00 UTC 2012
BCR-signalling synergizes with TLR-signalling for induction of AID and immunoglobulin class-switching through the non-canonical NF-κB pathway.
Nature communications
Pone EJ,Zhang J,Mai T,White CA,Li G,Sakakura JK,Patel PJ,Al-Qahtani A,Zan H,Xu Z,Casali P
13-0114 was used in Magnetic cell separation to investigate the role of BCR-signalling in class switching DNA recombination.
Tue Apr 03 00:00:00 UTC 2012
Transcription factor Smad-independent T helper 17 cell induction by transforming-growth factor-β is mediated by suppression of eomesodermin.
Immunity
Ichiyama K,Sekiya T,Inoue N,Tamiya T,Kashiwagi I,Kimura A,Morita R,Muto G,Shichita T,Takahashi R,Yoshimura A
13-0114 was used in Magnetic cell separation to investigate the molecular mechanisms by which TGF-beta allows Th17 cell differentiation via Smad-independent mechanisms, showing that Eomesodermin is suppressed by TGF-beta via the JNK pathway allowing differentiation.
Fri May 27 00:00:00 UTC 2011
Emergency granulopoiesis promotes neutrophil-dendritic cell encounters that prevent mouse lung allograft acceptance.
Blood
Kreisel D,Sugimoto S,Zhu J,Nava R,Li W,Okazaki M,Yamamoto S,Ibrahim M,Huang HJ,Toth KA,Ritter JH,Krupnick AS,Miller MJ,Gelman AE
13-0114 was used in Magnetic cell separation to identify granulopoiesis-mediated augmentation of alloimmunity as a novel link between innate and adaptive immune responses after organ transplantation.
Thu Dec 01 00:00:00 UTC 2011
Abrogation of CD30 and OX40 signals prevents autoimmune disease in FoxP3-deficient mice.
The Journal of experimental medicine
Gaspal F,Withers D,Saini M,Bekiaris V,McConnell FM,White A,Khan M,Yagita H,Walker LS,Anderson G,Lane PJ
13-0114 was used in Immunofluorescence to investigate how blocking antibodies binding to OX40 and CD30 ligands also abrogated disease mediated by FoxP3-deficient T cells.
Mon Aug 01 00:00:00 UTC 2011
Magnetic-based purification of untouched mouse germinal center B cells for ex vivo manipulation and biochemical analysis.
Nature protocols
Cato MH,Yau IW,Rickert RC
13-0114 was used in Magnetic cell separation to provide a protocol for a novel magnetic bead separation scheme for the purification of mature B cells for biochemical assessment.
Thu Jun 09 00:00:00 UTC 2011
Defective hematopoietic stem cell and lymphoid progenitor development in the Ts65Dn mouse model of Down syndrome: potential role of oxidative stress.
Antioxidants & redox signaling
Lorenzo LP,Chen H,Shatynski KE,Clark S,Yuan R,Harrison DE,Yarowsky PJ,Williams MS
13-0114 was used in Flow cytometry/Cell sorting to identify a link between oxidative stress and diminished IL-7Rα expression and function associated with defective haematologic development in a DS mouse model.
Sat Oct 15 00:00:00 UTC 2011
A helminth immunomodulator exploits host signaling events to regulate cytokine production in macrophages.
PLoS pathogens
Klotz C,Ziegler T,Figueiredo AS,Rausch S,Hepworth MR,Obsivac N,Sers C,Lang R,Hammerstein P,Lucius R,Hartmann S
13-0114 was used in Magnetic cell separation to elucidate the signalling events in macrophages triggered by helminth immunomodulator cystatin, showing cytokine signalling is regulated in these macrophages.
Thu Jan 06 00:00:00 UTC 2011
Hepatic stellate cells function as regulatory bystanders.
Journal of immunology (Baltimore, Md. : 1950)
Ichikawa S,Mucida D,Tyznik AJ,Kronenberg M,Cheroutre H
13-0114 was used in Magnetic cell separation to suggest that HSC can function as regulatory bystanders, and therefore, by promoting Tregs and suppressing Th17 differentiation, they might represent key players in the mechanism that drives liver-induced tolerance.
Sun May 15 00:00:00 UTC 2011
IL-1β and TNFα-initiated IL-6-STAT3 pathway is critical in mediating inflammatory cytokines and RANKL expression in inflammatory arthritis.
International immunology
Mori T,Miyamoto T,Yoshida H,Asakawa M,Kawasumi M,Kobayashi T,Morioka H,Chiba K,Toyama Y,Yoshimura A
13-0114 was used in Functional assays to elucidate the role of cytokines in chronic inflammation and joint destruction in rheumatoid arthritis.
Tue Nov 01 00:00:00 UTC 2011
PAD4-mediated neutrophil extracellular trap formation is not required for immunity against influenza infection.
PloS one
Hemmers S,Teijaro JR,Arandjelovic S,Mowen KA
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Nov 15 00:00:00 UTC 2011
Basophils orchestrate chronic allergic dermatitis and protective immunity against helminths.
Immunity
Ohnmacht C,Schwartz C,Panzer M,Schiedewitz I,Naumann R,Voehringer D
13-0114 was used in Flow cytometry/Cell sorting to demonstrate that basophils play an important role for protective immunity against helminths and orchestrate chronic allergic inflammation.
Fri Sep 24 00:00:00 UTC 2010
Moderate oxygen augments lipopolysaccharide-induced lung injury in mice.
American journal of physiology. Lung cellular and molecular physiology
Aggarwal NR,D'Alessio FR,Tsushima K,Files DC,Damarla M,Sidhaye VK,Fraig MM,Polotsky VY,King LS
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Mar 01 00:00:00 UTC 2010
Lack of interleukin-12 in p40-deficient mice leads to poor CD8+ T-cell immunity against Encephalitozoon cuniculi infection.
Infection and immunity
Moretto MM,Lawlor EM,Khan IA
13-0114 was used in Magnetic cell separation to demonstrate the important role of IL-12 in inducing an effective CD8+ T cell response against Encephalitozoon cuniculi Infection.
Tue Jun 01 00:00:00 UTC 2010
Expression of the chemokine binding protein M3 promotes marked changes in the accumulation of specific leukocytes subsets within the intestine.
Gastroenterology
Shang L,Thirunarayanan N,Viejo-Borbolla A,Martin AP,Bogunovic M,Marchesi F,Unkeless JC,Ho Y,Furtado GC,Alcami A,Merad M,Mayer L,Lira SA
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2009
RAG-1 and ATM coordinate monoallelic recombination and nuclear positioning of immunoglobulin loci.
Nature immunology
Hewitt SL,Yin B,Ji Y,Chaumeil J,Marszalek K,Tenthorey J,Salvagiotto G,Steinel N,Ramsey LB,Ghysdael J,Farrar MA,Sleckman BP,Schatz DG,Busslinger M,Bassing CH,Skok JA
13-0114 was used in Flow cytometry/Cell sorting to investigate whether coordinated recombination of homologous antigen receptor loci is important for allelic exclusion.
Mon Jun 01 00:00:00 UTC 2009
Development of immunoglobulin lambda-chain-positive B cells, but not editing of immunoglobulin kappa-chain, depends on NF-kappaB signals.
Nature immunology
Derudder E,Cadera EJ,Vahl JC,Wang J,Fox CJ,Zha S,van Loo G,Pasparakis M,Schlissel MS,Schmidt-Supprian M,Rajewsky K
13-0114 was used in Magnetic cell separation to define two distinct, consecutive phases of early B cell development that differ in their dependence on IKK-mediated NF-kappaB signaling.
Mon Jun 01 00:00:00 UTC 2009
A murine DC-SIGN homologue contributes to early host defense against Mycobacterium tuberculosis.
The Journal of experimental medicine
Tanne A,Ma B,Boudou F,Tailleux L,Botella H,Badell E,Levillain F,Taylor ME,Drickamer K,Nigou J,Dobos KM,Puzo G,Vestweber D,Wild MK,Marcinko M,Sobieszczuk P,Stewart L,Lebus D,Gicquel B,Neyrolles O
13-0114 was used in Immunohistochemistry on paraffin embedded tissues to investigate the function of DC-SIGN in the host response to pathogens in vivo.
Mon Sep 28 00:00:00 UTC 2009
Tissue-resident macrophages protect the liver from ischemia reperfusion injury via a heme oxygenase-1-dependent mechanism.
Molecular therapy : the journal of the American Society of Gene Therapy
Devey L,Ferenbach D,Mohr E,Sangster K,Bellamy CO,Hughes J,Wigmore SJ
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2009
Dendritic cells are required for optimal activation of natural killer functions following primary infection with herpes simplex virus type 1.
Journal of virology
Kassim SH,Rajasagi NK,Ritz BW,Pruett SB,Gardner EM,Chervenak R,Jennings SR
13-0114 was used in Flow cytometry/Cell sorting to provide evidence that functional DCs are required for optimal IFN-gamma expression and cytolytic function by NK cells following HSV-1 infection.
Wed Apr 01 00:00:00 UTC 2009
Combination of all-trans retinoic acid and a human papillomavirus therapeutic vaccine suppresses the number and function of immature myeloid cells and enhances antitumor immunity.
Cancer science
Song X,Ye D,Liu B,Cui J,Zhao X,Yi L,Liang J,Song J,Zhang Z,Zhao Q
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Sun Feb 01 00:00:00 UTC 2009
Attenuation of AD-like neuropathology by harnessing peripheral immune cells: local elevation of IL-10 and MMP-9.
Journal of neurochemistry
Koronyo-Hamaoui M,Ko MK,Koronyo Y,Azoulay D,Seksenyan A,Kunis G,Pham M,Bakhsheshian J,Rogeri P,Black KL,Farkas DL,Schwartz M
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Tue Dec 01 00:00:00 UTC 2009
The survival of memory CD4+ T cells within the gut lamina propria requires OX40 and CD30 signals.
Journal of immunology (Baltimore, Md. : 1950)
Withers DR,Jaensson E,Gaspal F,McConnell FM,Eksteen B,Anderson G,Agace WW,Lane PJL
13-0114 was used in Immunofluorescence to investigate how signals through CD30 and OX40 may be required for the survival of CD4+ T cells within the gut lamina propria.
Thu Oct 15 00:00:00 UTC 2009
Leptin receptor is expressed in thymus medulla and leptin protects against thymic remodeling during endotoxemia-induced thymus involution.
The Journal of endocrinology
Gruver AL,Ventevogel MS,Sempowski GD
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Thu Oct 01 00:00:00 UTC 2009
TLR2 and its co-receptors determine responses of macrophages and dendritic cells to lipoproteins of Mycobacterium tuberculosis.
Cellular immunology
Drage MG,Pecora ND,Hise AG,Febbraio M,Silverstein RL,Golenbock DT,Boom WH,Harding CV
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jun 12 00:00:00 UTC 2009
Mycobacterium bovis BCG decreases MHC-II expression in vivo on murine lung macrophages and dendritic cells during aerosol infection.
Cellular immunology
Pecora ND,Fulton SA,Reba SM,Drage MG,Simmons DP,Urankar-Nagy NJ,Boom WH,Harding CV
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Fri Jan 02 00:00:00 UTC 2009
Epidermal Langerhans cells promote skin allograft rejection in mice with NF-kappa B-impaired T cells.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Molinero LL,Zhou P,Wang Y,Harlin H,Kee B,Abraham C,Alegre ML
13-0114 was used in Flow cytometry/Cell sorting to suggest that Langerhans cells allow NF-κB-impaired T-cells to reach an activation threshold sufficient for skin allograft rejection.
Tue Jan 01 00:00:00 UTC 2008
Dynamics of lung macrophage activation in response to helminth infection.
Journal of leukocyte biology
Siracusa MC,Reece JJ,Urban JF,Scott AL
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Immunofluorescence
Mon Dec 01 00:00:00 UTC 2008
Impaired T-cell priming in vivo resulting from dysfunction of WASp-deficient dendritic cells.
Blood
Bouma G,Burns S,Thrasher AJ
13-0114 was used in Immunohistochemistry on frozen tissues to investigate the contribution of dendritic cell dysfunction to immune dysregulation in Wiskott-Aldrich syndrome.
Sat Dec 15 00:00:00 UTC 2007
Loss of T cell receptor-induced Bmi-1 in the KLRG1(+) senescent CD8(+) T lymphocyte.
Proceedings of the National Academy of Sciences of the United States of America
Heffner M,Fearon DT
13-0114 was used in Immunopurification to indicate that Bmi-1 is required for CD8(+) T cell clonal expansion and is positively regulated by receptors that mediate this response.
Tue Aug 14 00:00:00 UTC 2007
Murine CXCL14 is dispensable for dendritic cell function and localization within peripheral tissues.
Molecular and cellular biology
Meuter S,Schaerli P,Roos RS,Brandau O,Bösl MR,von Andrian UH,Moser B
13-0114 was used in Immunohistochemistry on frozen tissues to investigate the process of dendritic cell recruitment to peripheral tissues, showing that murine CXCL14 is dispensable for dendritic cell function and localisation.
Thu Feb 01 00:00:00 UTC 2007
Role of CD80 and CD86 in host immune responses to the recombinant hemagglutinin domain of Porphyromonas gingivalis gingipain and in the adjuvanticity of cholera toxin B and monophosphoryl lipid A.
Vaccine
Zhang P,Lewis JP,Michalek SM,Katz J
13-0114 was used in Flow cytometry/Cell sorting to demonstrate CD80 and CD86 play a role in mediating a systemic immune response towards P. gingivalis proteases.
Tue Aug 14 00:00:00 UTC 2007
The mouse mammary carcinoma 4T1: characterization of the cellular landscape of primary tumours and metastatic tumour foci.
International journal of experimental pathology
DuPré SA,Redelman D,Hunter KW
Published figure using CD11c monoclonal antibody (Product # 13-0114-82) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2007
Characterization of Siglec-H as a novel endocytic receptor expressed on murine plasmacytoid dendritic cell precursors.
Blood
Zhang J,Raper A,Sugita N,Hingorani R,Salio M,Palmowski MJ,Cerundolo V,Crocker PR
13-0114 was used in Flow cytometry/Cell sorting to analyse the cellular expression pattern and functional properties of Siglec-H.
Mon May 01 00:00:00 UTC 2006
Essential roles of DC-derived IL-15 as a mediator of inflammatory responses in vivo.
The Journal of experimental medicine
Ohteki T,Tada H,Ishida K,Sato T,Maki C,Yamada T,Hamuro J,Koyasu S
Mon Oct 02 00:00:00 UTC 2006
Essential roles of DC-derived IL-15 as a mediator of inflammatory responses in vivo.
The Journal of experimental medicine
Ohteki T,Tada H,Ishida K,Sato T,Maki C,Yamada T,Hamuro J,Koyasu S
Mon Oct 02 00:00:00 UTC 2006
Induction of robust immune responses against human immunodeficiency virus is supported by the inherent tropism of adeno-associated virus type 5 for dendritic cells.
Journal of virology
Xin KQ,Mizukami H,Urabe M,Toda Y,Shinoda K,Yoshida A,Oomura K,Kojima Y,Ichino M,Klinman D,Ozawa K,Okuda K
Fri Dec 01 00:00:00 UTC 2006
In vivo ablation of CD11c-positive dendritic cells increases susceptibility to herpes simplex virus type 1 infection and diminishes NK and T-cell responses.
Journal of virology
Kassim SH,Rajasagi NK,Zhao X,Chervenak R,Jennings SR
13-0114 was used in Flow cytometry/Cell sorting to investigate the role of different dendritic cell subsets in response to herpes simplex virus type 1 infection in mice.
Sat Apr 01 00:00:00 UTC 2006
The existence of CD11c+ sentinel and F4/80+ interstitial dendritic cells in dental pulp and their dynamics and functional properties.
International immunology
Zhang J,Kawashima N,Suda H,Nakano Y,Takano Y,Azuma M
Fri Sep 01 00:00:00 UTC 2006