Product References
IL-22 initiates an IL-18-dependent epithelial response circuit to enforce intestinal host defence.
Nature communications
Chiang HY,Lu HH,Sudhakar JN,Chen YW,Shih NS,Weng YT,Shui JW
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue Feb 15 00:00:00 EST 2022
FACS and immunomagnetic isolation of early erythroid progenitor cells from mouse fetal liver.
STAR protocols
Braun TW,Kuoch MK,Khandros E,Li H
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Fri Mar 18 00:00:00 EDT 2022
Hormone-Responsive BMP Signaling Expands Myoepithelial Cell Lineages and Prevents Alveolar Precocity in Mammary Gland.
Frontiers in cell and developmental biology
Shao C,Lou P,Liu R,Bi X,Li G,Yang X,Sheng X,Xu J,Lv C,Yu Z
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue Aug 03 00:00:00 EDT 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Plasmacytoid Dendritic Cells and Type I Interferon Promote Extrafollicular B Cell Responses to Extracellular Self-DNA.
Immunity
Soni C,Perez OA,Voss WN,Pucella JN,Serpas L,Mehl J,Ching KL,Goike J,Georgiou G,Ippolito GC,Sisirak V,Reizis B
13-0242-82 was used in Flow Cytometry to establish extrafollicular B cell differentiation into short-lived AFCs as a key mechanism of anti-DNA autoreactivity and reveal a major contribution of pDCs, endosomal Toll-like receptors (TLRs), and IFN-I to this pathway.
Tue Jun 16 00:00:00 EDT 2020
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Transcriptomic and proteomic signatures of stemness and differentiation in the colon crypt.
Communications biology
Habowski AN,Flesher JL,Bates JM,Tsai CF,Martin K,Zhao R,Ganesan AK,Edwards RA,Shi T,Wiley HS,Shi Y,Hertel KJ,Waterman ML
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Wed Aug 19 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
An Nfil3-Zeb2-Id2 pathway imposes Irf8 enhancer switching during cDC1 development.
Nature immunology
Bagadia P,Huang X,Liu TT,Durai V,Grajales-Reyes GE,Nitschké M,Modrusan Z,Granja JM,Satpathy AT,Briseño CG,Gargaro M,Iwata A,Kim S,Chang HY,Shaw AS,Murphy TL,Murphy KM
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Lifelong n-3 Polyunsaturated Fatty Acid Exposure Modulates Size of Mammary Epithelial Cell Populations and Expression of Caveolae Resident Proteins in Fat-1 Mice.
Nutrients
Hillyer LM,Kang JX,Ma DWL
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue Oct 15 00:00:00 EDT 2019
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Wnt Effector TCF4 Is Dispensable for Wnt Signaling in Human Cancer Cells.
Genes
Hrckulak D,Janeckova L,Lanikova L,Kriz V,Horazna M,Babosova O,Vojtechova M,Galuskova K,Sloncova E,Korinek V
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
A Paradoxical Tumor-Suppressor Role for the Rac1 Exchange Factor Vav1 in T Cell Acute Lymphoblastic Leukemia.
Cancer cell
Robles-Valero J,Lorenzo-Martín LF,Menacho-Márquez M,Fernández-Pisonero I,Abad A,Camós M,Toribio ML,Espinosa L,Bigas A,Bustelo XR
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Mon Nov 13 00:00:00 EST 2017
Adipocyte Accumulation in the Bone Marrow during Obesity and Aging Impairs Stem Cell-Based Hematopoietic and Bone Regeneration.
Cell stem cell
Ambrosi TH,Scialdone A,Graja A,Gohlke S,Jank AM,Bocian C,Woelk L,Fan H,Logan DW,Schürmann A,Saraiva LR,Schulz TJ
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Metabolic shifts in residual breast cancer drive tumor recurrence.
The Journal of clinical investigation
Havas KM,Milchevskaya V,Radic K,Alladin A,Kafkia E,Garcia M,Stolte J,Klaus B,Rotmensz N,Gibson TJ,Burwinkel B,Schneeweiss A,Pruneri G,Patil KR,Sotillo R,Jechlinger M
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
CD24 induces changes to the surface receptors of B cell microvesicles with variable effects on their RNA and protein cargo.
Scientific reports
Ayre DC,Chute IC,Joy AP,Barnett DA,Hogan AM,Grüll MP,Peña-Castillo L,Lang AS,Lewis SM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in T-Cell Activation
Thu Aug 17 00:00:00 EDT 2017
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Immunofluorescence
Tue Dec 20 00:00:00 EST 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Immunofluorescence
Tue Dec 20 00:00:00 EST 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Immunofluorescence
Tue Dec 20 00:00:00 EST 2016
Rapid depot-specific activation of adipocyte precursor cells at the onset of obesity.
Nature cell biology
Jeffery E,Church CD,Holtrup B,Colman L,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
The immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Immunity & ageing : I & A
Matsuda T,Yanase S,Takaoka A,Maruyama M
13-0242 was used in Flow cytometry/Cell sorting to show the immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Mon Mar 02 00:00:00 EST 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
A critical role for the regulated wnt-myc pathway in naive T cell survival.
Journal of immunology (Baltimore, Md. : 1950)
Wong C,Chen C,Wu Q,Liu Y,Zheng P
13-0242 was used in Magnetic cell separation to investigate the role of Wnt signalling in mature naive T cells.
Thu Jan 01 00:00:00 EST 2015
Dynamic upregulation of CD24 in pre-adipocytes promotes adipogenesis.
Adipocyte
Smith NC,Fairbridge NA,Pallegar NK,Christian SL
13-0242 was used in Flow cytometry/Cell sorting to investigate the role and regulation of CD24 during adipogenesis, showing that dynamic upregulation of CD24 in pre-adipocytes promotes adipogenesis.
Mon Jul 13 00:00:00 EDT 2015
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Fri Feb 06 00:00:00 EST 2015
Ebf2 is a selective marker of brown and beige adipogenic precursor cells.
Proceedings of the National Academy of Sciences of the United States of America
Wang W,Kissig M,Rajakumari S,Huang L,Lim HW,Won KJ,Seale P
13-0242 was used in Flow cytometry/Cell sorting to study Ebf2 specifically marking and regulating the molecular profile of brown preadipose cells.
Tue Oct 07 00:00:00 EDT 2014
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
13-0242 was used in Flow cytometry/Cell sorting to demonstrate that RhoF promotes MZ B cell development in the spleen.
Fri Aug 01 00:00:00 EDT 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
13-0242 was used in Flow cytometry/Cell sorting to demonstrate that RhoF promotes MZ B cell development in the spleen.
Fri Aug 01 00:00:00 EDT 2014
Prospective identification of functionally distinct stem cells and neurosphere-initiating cells in adult mouse forebrain.
eLife
Mich JK,Signer RA,Nakada D,Pineda A,Burgess RJ,Vue TY,Johnson JE,Morrison SJ
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Wed May 07 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
Characterization of the adipocyte cellular lineage in vivo.
Nature cell biology
Berry R,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
13-0242 was used in Flow cytometry/Cell sorting to provide data to show the existence of multi-potent mammary progenitor cells that have pregnancy-specific function.
Mon Dec 08 00:00:00 EST 2014
FLT3-ITDs instruct a myeloid differentiation and transformation bias in lymphomyeloid multipotent progenitors.
Cell reports
Mead AJ,Kharazi S,Atkinson D,Macaulay I,Pecquet C,Loughran S,Lutteropp M,Woll P,Chowdhury O,Luc S,Buza-Vidas N,Ferry H,Clark SA,Goardon N,Vyas P,Constantinescu SN,Sitnicka E,Nerlov C,Jacobsen SE
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Thu Jun 27 00:00:00 EDT 2013
Lunatic fringe deficiency cooperates with the Met/Caveolin gene amplicon to induce basal-like breast cancer.
Cancer cell
Xu K,Usary J,Kousis PC,Prat A,Wang DY,Adams JR,Wang W,Loch AJ,Deng T,Zhao W,Cardiff RD,Yoon K,Gaiano N,Ling V,Beyene J,Zacksenhaus E,Gridley T,Leong WL,Guidos CJ,Perou CM,Egan SE
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue May 15 00:00:00 EDT 2012
Restricted expression of new guanine nucleotide exchange factor Zizimin2 in aged acquired immune system.
Nagoya journal of medical science
Jia Y,Sakabe I,Matsuda T,Hayakawa T,Maruyama M
13-0242 was used in Flow cytometry/Cell sorting to elucidate the molecular mechanism of age-related decline of immune systems.
Wed Aug 01 00:00:00 EDT 2012
Regulation of TCRβ allelic exclusion by gene segment proximity and accessibility.
Journal of immunology (Baltimore, Md. : 1950)
Kondilis-Mangum HD,Shih HY,Mahowald G,Sleckman BP,Krangel MS
13-0242 was used in Flow cytometry/Cell sorting to analyse the contributions of chromatin accessibility and locus conformation to feedback inhibition.
Thu Dec 15 00:00:00 EST 2011
Stem cell marker prominin-1 regulates branching morphogenesis, but not regenerative capacity, in the mammary gland.
Developmental dynamics : an official publication of the American Association of Anatomists
Anderson LH,Boulanger CA,Smith GH,Carmeliet P,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2011
A multifunctional 3D co-culture system for studies of mammary tissue morphogenesis and stem cell biology.
PloS one
Campbell JJ,Davidenko N,Caffarel MM,Cameron RE,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Flow Cytometry
Mon Mar 05 00:00:00 EST 2012
Tumor-initiating function of nucleostemin-enriched mammary tumor cells.
Cancer research
Lin T,Meng L,Li Y,Tsai RY
13-0242 was used in Flow cytometry/Cell sorting to establish features of NS-enriched mammary tumour cells to determine their potential as a therapeutic target.
Mon Nov 15 00:00:00 EST 2010
Robust in vivo transduction of nervous system and neural stem cells by early gestational intra amniotic gene transfer using lentiviral vector.
Molecular therapy : the journal of the American Society of Gene Therapy
Stitelman DH,Endo M,Bora A,Muvarak N,Zoltick PW,Flake AW,Brazelton TR
Published figure using CD24 monoclonal antibody (Product # 13-0242-82) in Immunofluorescence
Wed Sep 01 00:00:00 EDT 2010
Expanded CD23(+)/CD21(hi) B cells in inflamed lymph nodes are associated with the onset of inflammatory-erosive arthritis in TNF-transgenic mice and are targets of anti-CD20 therapy.
Journal of immunology (Baltimore, Md. : 1950)
Li J,Kuzin I,Moshkani S,Proulx ST,Xing L,Skrombolas D,Dunn R,Sanz I,Schwarz EM,Bottaro A
13-0242 was used in Flow cytometry/Cell sorting to investigate the pathogenic role of B lymphocytes in rheumatoid arthritis, showing that CD23(+)/CD21(hi) B cells in inflamed lymph nodes are associated with the onset of inflammatory-erosive arthritis.
Tue Jun 01 00:00:00 EDT 2010
MAP kinase phosphatase activity sets the threshold for thymocyte positive selection.
Proceedings of the National Academy of Sciences of the United States of America
Bettini ML,Kersh GJ
13-0242 was used in Flow cytometry/Cell sorting to demonstrate that MKP activity controls thymocyte cell fate by regulating the TCR signaling threshold that induces positive selection.
Tue Oct 09 00:00:00 EDT 2007