Product References
Mesenchymal Stem Cell-Derived Exosomes Attenuate Murine Cytomegalovirus-Infected Pneumonia via NF-κB/NLRP3 Signaling Pathway.
Viruses
Chen F,Chen Z,Wu HT,Chen XX,Zhan P,Wei ZY,Ouyang Z,Jiang X,Shen A,Luo MH,Liu Q,Zhou YP,Qin A
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Tue Apr 16 00:00:00 UTC 2024
Overexpression of fibroblast growth factor receptor 2 in bone marrow mesenchymal stem cells enhances osteogenesis and promotes critical cranial bone defect regeneration.
Frontiers in cell and developmental biology
Zhou Y,Zhu P,Shen S,Wang Y,Li B,Guo B,Li H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Sun Jun 04 00:00:00 UTC 2023
Fetal Muse-based therapy prevents lethal radio-induced gastrointestinal syndrome by intestinal regeneration.
Stem cell research & therapy
Dushime H,Moreno SG,Linard C,Adrait A,Couté Y,Peltzer J,Messiaen S,Torres C,Bensemmane L,Lewandowski D,Romeo PH,Petit V,Gault N
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Fri Aug 11 00:00:00 UTC 2023
Age-related ultrastructural changes in spheroids of the adipose-derived multipotent mesenchymal stromal cells from ovariectomized mice.
Frontiers in cellular neuroscience
Kyryk V,Tsupykov O,Ustymenko A,Smozhanik E,Govbakh I,Butenko G,Skibo G
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Tue Mar 07 00:00:00 UTC 2023
Hypoxia pretreatment enhances the therapeutic potential of mesenchymal stem cells (BMSCs) on ozone-induced lung injury in rats.
Cell and tissue research
Abdelrahman SA,Abdelrahman AA,Samy W,Dessouky AA,Ahmed SM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunohistochemistry
Mon Aug 01 00:00:00 UTC 2022
Induction of mesenchymal stem cell‑like transformation in rat primary glial cells using hypoxia, mild hypothermia and growth factors.
Molecular medicine reports
Wei H,Zhou W,Hu G,Shi C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Extracellular vesicles from recombinant cell factories improve the activity and efficacy of enzymes defective in lysosomal storage disorders.
Journal of extracellular vesicles
Seras-Franzoso J,Díaz-Riascos ZV,Corchero JL,González P,García-Aranda N,Mandaña M,Riera R,Boullosa A,Mancilla S,Grayston A,Moltó-Abad M,Garcia-Fruitós E,Mendoza R,Pintos-Morell G,Albertazzi L,Rosell A,Casas J,Villaverde A,Schwartz S,Abasolo I
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Mon Mar 01 00:00:00 UTC 2021
Distinct skeletal stem cell types orchestrate long bone skeletogenesis.
eLife
Ambrosi TH,Sinha R,Steininger HM,Hoover MY,Murphy MP,Koepke LS,Wang Y,Lu WJ,Morri M,Neff NF,Weissman IL,Longaker MT,Chan CK
13-1051-82 was used in Flow Cytometry to address previously unappreciated shortcomings of SSC research.
Mon Jul 19 00:00:00 UTC 2021
C/EBPα and GATA-2 Mutations Induce Bilineage Acute Erythroid Leukemia through Transformation of a Neomorphic Neutrophil-Erythroid Progenitor.
Cancer cell
Di Genua C,Valletta S,Buono M,Stoilova B,Sweeney C,Rodriguez-Meira A,Grover A,Drissen R,Meng Y,Beveridge R,Aboukhalil Z,Karamitros D,Belderbos ME,Bystrykh L,Thongjuea S,Vyas P,Nerlov C
13-1051 was used in Flow cytometry/Cell sorting to show that combined biallelic Cebpa and Gata2 zinc finger-1 (ZnF1) mutations cooperatively induce bilineage AEL, and that the major leukemia-initiating cell (LIC) population has a neutrophil-monocyte progenitor (NMP) phenotype.
Mon May 11 00:00:00 UTC 2020
Extracellular HMGB-1 activates inflammatory signaling in tendon cells and tissues.
Therapeutic advances in chronic disease
Zhang C,Gu X,Zhao G,Wang W,Shao J,Zhu J,Yuan T,Sun J,Nie D,Zhou Y
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Sun Apr 17 00:00:00 UTC 2022
Targeting compensatory MEK/ERK activation increases JAK inhibitor efficacy in myeloproliferative neoplasms.
The Journal of clinical investigation
Stivala S,Codilupi T,Brkic S,Baerenwaldt A,Ghosh N,Hao-Shen H,Dirnhofer S,Dettmer MS,Simillion C,Kaufmann BA,Chiu S,Keller M,Kleppe M,Hilpert M,Buser AS,Passweg JR,Radimerski T,Skoda RC,Levine RL,Meyer SC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Mon Mar 04 00:00:00 UTC 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
The microRNAs miR-204 and miR-211 maintain joint homeostasis and protect against osteoarthritis progression.
Nature communications
Huang J,Zhao L,Fan Y,Liao L,Ma PX,Xiao G,Chen D
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Fri Jun 28 00:00:00 UTC 2019
Isolation and functional assessment of mouse skeletal stem cell lineage.
Nature protocols
Gulati GS,Murphy MP,Marecic O,Lopez M,Brewer RE,Koepke LS,Manjunath A,Ransom RC,Salhotra A,Weissman IL,Longaker MT,Chan CKF
13-1051-82 was used in flow cytometry to describe a flow cytometry-based approach that does not require flow cytometry selection and that uses eight surface markers to distinguish and isolate mouse skeletal stem cells (mSSCs); bone, cartilage, and stromal progenitors (mBCSPs); and five downstream differentiated subtypes, including chondroprogenitors, two types of osteoprogenitors, and two types of hematopoiesis-supportive stroma.
Fri Jun 01 00:00:00 UTC 2018
Isolation and functional assessment of mouse skeletal stem cell lineage.
Nature protocols
Gulati GS,Murphy MP,Marecic O,Lopez M,Brewer RE,Koepke LS,Manjunath A,Ransom RC,Salhotra A,Weissman IL,Longaker MT,Chan CKF
13-1051-82 was used in flow cytometry to describe a flow cytometry-based approach that does not require flow cytometry selection and that uses eight surface markers to distinguish and isolate mouse skeletal stem cells (mSSCs); bone, cartilage, and stromal progenitors (mBCSPs); and five downstream differentiated subtypes, including chondroprogenitors, two types of osteoprogenitors, and two types of hematopoiesis-supportive stroma.
Fri Jun 01 00:00:00 UTC 2018
Utilizing Confocal Microscopy to Characterize Human and Mouse Adipose Tissue.
Tissue engineering. Part C, Methods
Blackshear CP,Borrelli MR,Shen EZ,Ransom RC,Chung NN,Vistnes SM,Irizarry D,Nazerali R,Momeni A,Longaker MT,Wan DC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunohistochemistry
Mon Oct 01 00:00:00 UTC 2018
Utilizing Confocal Microscopy to Characterize Human and Mouse Adipose Tissue.
Tissue engineering. Part C, Methods
Blackshear CP,Borrelli MR,Shen EZ,Ransom RC,Chung NN,Vistnes SM,Irizarry D,Nazerali R,Momeni A,Longaker MT,Wan DC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunohistochemistry
Mon Oct 01 00:00:00 UTC 2018
RhoA, Rac1, and Cdc42 differentially regulate αSMA and collagen I expression in mesenchymal stem cells.
The Journal of biological chemistry
Ge J,Burnier L,Adamopoulou M,Kwa MQ,Schaks M,Rottner K,Brakebusch C
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Fri Jun 15 00:00:00 UTC 2018
Small non-coding RNA expression in mouse nephrogenic mesenchymal progenitors.
Scientific data
Phua YL,Clugston A,Chen KH,Kostka D,Ho J
13-1051-82 was used in Magnetic cell separation to characterise the microRNA profile of mouse nephrogenic mesenchymal progenitors.
Tue Nov 13 00:00:00 UTC 2018
Endoglin prevents vascular malformation by regulating flow-induced cell migration and specification through VEGFR2 signalling.
Nature cell biology
Jin Y,Muhl L,Burmakin M,Wang Y,Duchez AC,Betsholtz C,Arthur HM,Jakobsson L
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunofluorescence
Thu Jun 01 00:00:00 UTC 2017
Cardiosphere-Derived Cells Require Endoglin for Paracrine-Mediated Angiogenesis.
Stem cell reports
Redgrave RE,Tual-Chalot S,Davison BJ,Singh E,Hall D,Amirrasouli MM,Gilchrist D,Medvinsky A,Arthur HM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunofluorescence
Tue May 09 00:00:00 UTC 2017
Cardiosphere-Derived Cells Require Endoglin for Paracrine-Mediated Angiogenesis.
Stem cell reports
Redgrave RE,Tual-Chalot S,Davison BJ,Singh E,Hall D,Amirrasouli MM,Gilchrist D,Medvinsky A,Arthur HM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunofluorescence
Tue May 09 00:00:00 UTC 2017
Mice Lacking Endoglin in Macrophages Show an Impaired Immune Response.
PLoS genetics
Ojeda-Fernández L,Recio-Poveda L,Aristorena M,Lastres P,Blanco FJ,Sanz-Rodríguez F,Gallardo-Vara E,de las Casas-Engel M,Corbí Á,Arthur HM,Bernabeu C,Botella LM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Mice Lacking Endoglin in Macrophages Show an Impaired Immune Response.
PLoS genetics
Ojeda-Fernández L,Recio-Poveda L,Aristorena M,Lastres P,Blanco FJ,Sanz-Rodríguez F,Gallardo-Vara E,de las Casas-Engel M,Corbí Á,Arthur HM,Bernabeu C,Botella LM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Hydrogen sulfide metabolism regulates endothelial solute barrier function.
Redox biology
Yuan S,Pardue S,Shen X,Alexander JS,Orr AW,Kevil CG
13-1051 was used in Flow cytometry/Cell sorting to demonstrate that endothelial solute permeability is critically regulated via exogenous and endogenous sulfide bioavailability with a prominent role of polysulfides.
Sat Oct 01 00:00:00 UTC 2016
KLF4-dependent phenotypic modulation of smooth muscle cells has a key role in atherosclerotic plaque pathogenesis.
Nature medicine
Shankman LS,Gomez D,Cherepanova OA,Salmon M,Alencar GF,Haskins RM,Swiatlowska P,Newman AA,Greene ES,Straub AC,Isakson B,Randolph GJ,Owens GK
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Anti-metastatic action of FAK inhibitor OXA-11 in combination with VEGFR-2 signaling blockade in pancreatic neuroendocrine tumors.
Clinical & experimental metastasis
Moen I,Gebre M,Alonso-Camino V,Chen D,Epstein D,McDonald DM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunofluorescence
Tue Dec 01 00:00:00 UTC 2015
Tie1 is required for lymphatic valve and collecting vessel development.
Developmental biology
Qu X,Zhou B,Scott Baldwin H
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Immunofluorescence
Sun Mar 01 00:00:00 UTC 2015
Endoglin potentiates nitric oxide synthesis to enhance definitive hematopoiesis.
Biology open
Nasrallah R,Knezevic K,Thai T,Thomas SR,Göttgens B,Lacaud G,Kouskoff V,Pimanda JE
13105182 was used in flow cytometry to investigate the role of endoglin in the development of hematopoietic cells.
Fri May 15 00:00:00 UTC 2015
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Thu Mar 13 00:00:00 UTC 2014
Niche-based screening identifies small-molecule inhibitors of leukemia stem cells.
Nature chemical biology
Hartwell KA,Miller PG,Mukherjee S,Kahn AR,Stewart AL,Logan DJ,Negri JM,Duvet M,Järås M,Puram R,Dancik V,Al-Shahrour F,Kindler T,Tothova Z,Chattopadhyay S,Hasaka T,Narayan R,Dai M,Huang C,Shterental S,Chu LP,Haydu JE,Shieh JH,Steensma DP,Munoz B,Bittker JA,Shamji AF,Clemons PA,Tolliday NJ,Carpenter AE,Gilliland DG,Stern AM,Moore MAS,Scadden DT,Schreiber SL,Ebert BL,Golub TR
13-1051 was used in Magnetic cell separation to illustrate the power of merging physiologically relevant models with high-throughput screening to develop more effective therapies for acute leukemia.
Sun Dec 01 00:00:00 UTC 2013
Isolation and characterization of novel murine epiphysis derived mesenchymal stem cells.
PloS one
Cheng CC,Lian WS,Hsiao FS,Liu IH,Lin SP,Lee YH,Chang CC,Xiao GY,Huang HY,Cheng CF,Cheng WT,Wu SC
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Fri Sep 07 00:00:00 UTC 2012
Adipose-derived stem cells promote lymphangiogenesis in response to VEGF-C stimulation or TGF-β1 inhibition.
Future oncology (London, England)
Yan A,Avraham T,Zampell JC,Haviv YS,Weitman E,Mehrara BJ
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2011
Expression and role of FLT3 in regulation of the earliest stage of normal granulocyte-monocyte progenitor development.
Blood
Böiers C,Buza-Vidas N,Jensen CT,Pronk CJ,Kharazi S,Wittmann L,Sitnicka E,Hultquist A,Jacobsen SE
13-1051 was used in Flow cytometry/Cell sorting to investigate the role of FLT3 in early granulocyte-monocyte progenitors (GMPs), showing that most GMPs are derived from FLT3-expressing progenitors.
Thu Jun 17 00:00:00 UTC 2010
Delayed enrichment of mesenchymal cells promotes cardiac lineage and calcium transient development.
Journal of molecular and cellular cardiology
Grajales L,García J,Banach K,Geenen DL
13-1051 was used in Flow cytometry/Cell sorting to hypothesize that cardiac lineage development in BM-MSC is related to cell passage, culture milieu, and enrichment for specific cell subtypes before and during differentiation.
Thu Apr 01 00:00:00 UTC 2010
Pharmacologic targeting of a stem/progenitor population in vivo is associated with enhanced bone regeneration in mice.
The Journal of clinical investigation
Mukherjee S,Raje N,Schoonmaker JA,Liu JC,Hideshima T,Wein MN,Jones DC,Vallet S,Bouxsein ML,Pozzi S,Chhetri S,Seo YD,Aronson JP,Patel C,Fulciniti M,Purton LE,Glimcher LH,Lian JB,Stein G,Anderson KC,Scadden DT
13-1051 was used in Magnetic cell separation to show that a tissue-resident adult stem cell population can be pharmacologically modified to promote a regenerative function in adult animals.
Fri Feb 01 00:00:00 UTC 2008
Neo-organoid of marrow mesenchymal stromal cells secreting interleukin-12 for breast cancer therapy.
Cancer research
Eliopoulos N,Francois M,Boivin MN,Martineau D,Galipeau J
13-1051 was used in Flow cytometry/Cell sorting to establish the feasibility of using gene-enhanced MSCs in a cell-based neo-organoid approach for cancer treatment.
Sun Jun 15 00:00:00 UTC 2008
Circadian mechanisms in murine and human bone marrow mesenchymal stem cells following dexamethasone exposure.
Bone
Wu X,Yu G,Parks H,Hebert T,Goh BC,Dietrich MA,Pelled G,Izadpanah R,Gazit D,Bunnell BA,Gimble JM
Published figure using CD105 (Endoglin) monoclonal antibody (Product # 13-1051-82) in Flow Cytometry
Thu May 01 00:00:00 UTC 2008
A novel role for PECAM-1 (CD31) in regulating haematopoietic progenitor cell compartmentalization between the peripheral blood and bone marrow.
PloS one
Ross EA,Freeman S,Zhao Y,Dhanjal TS,Ross EJ,Lax S,Ahmed Z,Hou TZ,Kalia N,Egginton S,Nash G,Watson SP,Frampton J,Buckley CD
13-1051 was used in Immunofluorescence on frozen tissues to investigate the role of PECAM-1 in the bone marrow microenvironment.
Wed Jun 04 00:00:00 UTC 2008
Monitoring response to anticancer therapy by targeting microbubbles to tumor vasculature.
Clinical cancer research : an official journal of the American Association for Cancer Research
Korpanty G,Carbon JG,Grayburn PA,Fleming JB,Brekken RA
Mon Jan 01 00:00:00 UTC 2007
A role for endoglin as a suppressor of malignancy during mouse skin carcinogenesis.
Cancer research
Pérez-Gómez E,Villa-Morales M,Santos J,Fernández-Piqueras J,Gamallo C,Dotor J,Bernabéu C,Quintanilla M
Thu Nov 01 00:00:00 UTC 2007
Allogeneic marrow stromal cells are immune rejected by MHC class I- and class II-mismatched recipient mice.
Blood
Eliopoulos N,Stagg J,Lejeune L,Pommey S,Galipeau J
13-1051 was used in Flow cytometry/Cell sorting to strongly suggest that marrow stromal cells are not intrinsically immunoprivileged and cannot serve as a "universal donor" in immunocompetent MHC-mismatched recipients.
Thu Dec 15 00:00:00 UTC 2005
Liver sinusoidal endothelial cells tolerize T cells across MHC barriers in mice.
Journal of immunology (Baltimore, Md. : 1950)
Onoe T,Ohdan H,Tokita D,Shishida M,Tanaka Y,Hara H,Zhou W,Ishiyama K,Mitsuta H,Ide K,Asahara T
Fri Jul 01 00:00:00 UTC 2005