Product References
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Genomic analysis of human brain metastases treated with stereotactic radiosurgery reveals unique signature based on treatment failure.
iScience
Shireman JM,White Q,Ni Z,Mohanty C,Cai Y,Zhao L,Agrawal N,Gonugunta N,Wang X,Mccarthy L,Kasulabada V,Pattnaik A,Ahmed AU,Miller J,Kulwin C,Cohen-Gadol A,Payner T,Lin CT,Savage JJ,Lane B,Shiue K,Kamer A,Shah M,Iyer G,Watson G,Kendziorski C,Dey M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunohistochemistry
Fri Apr 19 00:00:00 UTC 2024
Osimertinib Covalently Binds to CD34 and Eliminates Myeloid Leukemia Stem/Progenitor Cells.
Cancer research
Xia L,Liu JY,Yang MY,Zhang XH,Jiang Y,Yin QQ,Luo CH,Liu HC,Kang ZJ,Zhang CT,Gao BB,Zhou AW,Cai HY,Waller EK,Yan JS,Lu Y
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Celecoxib induces adipogenic differentiation of hemangioma-derived mesenchymal stem cells through the PPAR-γ pathway in vitro and in vivo.
Experimental and therapeutic medicine
Wang Y,Kong L,Sun B,Cui J,Shen W
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Wed Jun 01 00:00:00 UTC 2022
Blocking STAT3/5 through direct or upstream kinase targeting in leukemic cutaneous T-cell lymphoma.
EMBO molecular medicine
Sorger H,Dey S,Vieyra-Garcia PA,Pölöske D,Teufelberger AR,de Araujo ED,Sedighi A,Graf R,Spiegl B,Lazzeri I,Braun T,Garces de Los Fayos Alonso I,Schlederer M,Timelthaler G,Kodajova P,Pirker C,Surbek M,Machtinger M,Graier T,Perchthaler I,Pan Y,Fink-Puches R,Cerroni L,Ober J,Otte M,Albrecht JD,Tin G,Abdeldayem A,Manaswiyoungkul P,Olaoye OO,Metzelder ML,Orlova A,Berger W,Wobser M,Nicolay JP,André F,Nguyen VA,Neubauer HA,Fleck R,Merkel O,Herling M,Heitzer E,Gunning PT,Kenner L,Moriggl R,Wolf P
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Wed Dec 07 00:00:00 UTC 2022
A fluorogenic probe for predicting treatment response in non-small cell lung cancer with EGFR-activating mutations.
Nature communications
Deng H,Lei Q,Wang C,Wang Z,Chen H,Wang G,Yang N,Huang D,Yu Q,Yao M,Xiao X,Zhu G,Cheng C,Li Y,Li F,Tian P,Li W
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Mon Nov 14 00:00:00 UTC 2022
Human placental mesenchymal stem cells regulate inflammation via the NF‑κB signaling pathway.
Experimental and therapeutic medicine
Liu Y,Zhang X,Hu Y,Kang M,Wu Y,Wang Y,Deng C
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
Pan-cancer analyses reveal cancer-type-specific fungal ecologies and bacteriome interactions.
Cell
Narunsky-Haziza L,Sepich-Poore GD,Livyatan I,Asraf O,Martino C,Nejman D,Gavert N,Stajich JE,Amit G,González A,Wandro S,Perry G,Ariel R,Meltser A,Shaffer JP,Zhu Q,Balint-Lahat N,Barshack I,Dadiani M,Gal-Yam EN,Patel SP,Bashan A,Swafford AD,Pilpel Y,Knight R,Straussman R
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunohistochemistry
Thu Sep 29 00:00:00 UTC 2022
An omic and multidimensional spatial atlas from serial biopsies of an evolving metastatic breast cancer.
Cell reports. Medicine
Johnson BE,Creason AL,Stommel JM,Keck JM,Parmar S,Betts CB,Blucher A,Boniface C,Bucher E,Burlingame E,Camp T,Chin K,Eng J,Estabrook J,Feiler HS,Heskett MB,Hu Z,Kolodzie A,Kong BL,Labrie M,Lee J,Leyshock P,Mitri S,Patterson J,Riesterer JL,Sivagnanam S,Somers J,Sudar D,Thibault G,Weeder BR,Zheng C,Nan X,Thompson RF,Heiser LM,Spellman PT,Thomas G,Demir E,Chang YH,Coussens LM,Guimaraes AR,Corless C,Goecks J,Bergan R,Mitri Z,Mills GB,Gray JW
14-0459-82 was used in Flow Cytometry to present illustrative examples of how integrative analyses of data reveal potential mechanisms of response and resistance and suggest novel therapeutic vulnerabilities.
Tue Feb 15 00:00:00 UTC 2022
NASH limits anti-tumour surveillance in immunotherapy-treated HCC.
Nature
Pfister D,Núñez NG,Pinyol R,Govaere O,Pinter M,Szydlowska M,Gupta R,Qiu M,Deczkowska A,Weiner A,Müller F,Sinha A,Friebel E,Engleitner T,Lenggenhager D,Moncsek A,Heide D,Stirm K,Kosla J,Kotsiliti E,Leone V,Dudek M,Yousuf S,Inverso D,Singh I,Teijeiro A,Castet F,Montironi C,Haber PK,Tiniakos D,Bedossa P,Cockell S,Younes R,Vacca M,Marra F,Schattenberg JM,Allison M,Bugianesi E,Ratziu V,Pressiani T,D'Alessio A,Personeni N,Rimassa L,Daly AK,Scheiner B,Pomej K,Kirstein MM,Vogel A,Peck-Radosavljevic M,Hucke F,Finkelmeier F,Waidmann O,Trojan J,Schulze K,Wege H,Koch S,Weinmann A,Bueter M,Rössler F,Siebenhüner A,De Dosso S,Mallm JP,Umansky V,Jugold M,Luedde T,Schietinger A,Schirmacher P,Emu B,Augustin HG,Billeter A,Müller-Stich B,Kikuchi H,Duda DG,Kütting F,Waldschmidt DT,Ebert MP,Rahbari N,Mei HE,Schulz AR,Ringelhan M,Malek N,Spahn S,Bitzer M,Ruiz de Galarreta M,Lujambio A,Dufour JF,Marron TU,Kaseb A,Kudo M,Huang YH,Djouder N,Wolter K,Zender L,Marche PN,Decaens T,Pinato DJ,Rad R,Mertens JC,Weber A,Unger K,Meissner F,Roth S,Jilkova ZM,Claassen M,Anstee QM,Amit I,Knolle P,Becher B,Llovet JM,Heikenwalder M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
An immune-humanized patient-derived xenograft model of estrogen-independent, hormone receptor positive metastatic breast cancer.
Breast cancer research : BCR
Scherer SD,Riggio AI,Haroun F,DeRose YS,Ekiz HA,Fujita M,Toner J,Zhao L,Li Z,Oesterreich S,Samatar AA,Welm AL
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Sat Oct 30 00:00:00 UTC 2021
Direct Comparative Analyses of 10X Genomics Chromium and Smart-seq2.
Genomics, proteomics & bioinformatics
Wang X,He Y,Zhang Q,Ren X,Zhang Z
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Targeting USP47 overcomes tyrosine kinase inhibitor resistance and eradicates leukemia stem/progenitor cells in chronic myelogenous leukemia.
Nature communications
Lei H,Xu HZ,Shan HZ,Liu M,Lu Y,Fang ZX,Jin J,Jing B,Xiao XH,Gao SM,Gao FH,Xia L,Yang L,Liu LG,Wang WW,Liu CX,Tong Y,Wu YZ,Zheng JK,Chen GQ,Zhou L,Wu YL
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunohistochemistry
Mon Jan 04 00:00:00 UTC 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
Clinical Significance of Circulating Tumor Cell Induced Epithelial-Mesenchymal Transition in Patients with Metastatic Colorectal Cancer by Single-Cell RNA-Sequencing.
Cancers
Kozuka M,Battaglin F,Jayachandran P,Wang J,Arai H,Soni S,Zhang W,Hirai M,Matsusaka S,Lenz HJ
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunocytochemistry
Tue Sep 28 00:00:00 UTC 2021
In vitro co-culture of human intestinal organoids and lamina propria-derived CD4+ T cells.
STAR protocols
Schreurs RRCE,Baumdick ME,Drewniak A,Bunders MJ
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Jun 18 00:00:00 UTC 2021
Longitudinal clonal tracking in humanized mice reveals sustained polyclonal repopulation of gene-modified human-HSPC despite vector integration bias.
Stem cell research & therapy
Suryawanshi GW,Arokium H,Kim S,Khamaikawin W,Lin S,Shimizu S,Chupradit K,Lee Y,Xie Y,Guan X,Suryawanshi V,Presson AP,An DS,Chen ISY
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Oct 07 00:00:00 UTC 2021
Bispecific T-Cell Engaging Antibodies Against MUC16 Demonstrate Efficacy Against Ovarian Cancer in Monotherapy and in Combination With PD-1 and VEGF Inhibition.
Frontiers in immunology
Yeku OO,Rao TD,Laster I,Kononenko A,Purdon TJ,Wang P,Cui Z,Liu H,Brentjens RJ,Spriggs D
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Oct 21 00:00:00 UTC 2021
Persistent NF-κB activation in muscle stem cells induces proliferation-independent telomere shortening.
Cell reports
Tichy ED,Ma N,Sidibe D,Loro E,Kocan J,Chen DZ,Khurana TS,Hasty P,Mourkioti F
14-0459-82 was used in Flow cytometry/Cell sorting to investigate NF-kB signalling on muscle stem cells.
Tue May 11 00:00:00 UTC 2021
SARS-CoV-2 Spike Protein-Directed Monoclonal Antibodies May Ameliorate COVID-19 Complications in APECED Patients.
Frontiers in immunology
Ferré EMN,Schmitt MM,Ochoa S,Rosen LB,Shaw ER,Burbelo PD,Stoddard JL,Rampertaap S,DiMaggio T,Bergerson JRE,Rosenzweig SD,Notarangelo LD,Holland SM,Lionakis MS
14-0459-82 was used in Flow Cytometry to assess the safety and clinical effects of early treatment with bamlanivimab and etesevimab.
Fri Sep 17 00:00:00 UTC 2021
DKK2 blockage-mediated immunotherapy enhances anti-angiogenic therapy of Kras mutated colorectal cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Hu J,Wang Z,Chen Z,Li A,Sun J,Zheng M,Wu J,Shen T,Qiao J,Li L,Li B,Wu D,Xiao Q
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Microsieves for the detection of circulating tumor cells in leukapheresis product in non-small cell lung cancer patients.
Translational lung cancer research
Tamminga M,Oomens L,Hiltermann TJN,Andree KC,Tibbe A,Broekmaat J,Schuuring E,Terstappen LWMM,Groen HJM
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunocytochemistry
Sat Aug 01 00:00:00 UTC 2020
Evaluation of K18-hACE2 Mice as a Model of SARS-CoV-2 Infection.
The American journal of tropical medicine and hygiene
Moreau GB,Burgess SL,Sturek JM,Donlan AN,Petri WA,Mann BJ
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
The novel reversible LSD1 inhibitor SP-2577 promotes anti-tumor immunity in SWItch/Sucrose-NonFermentable (SWI/SNF) complex mutated ovarian cancer.
PloS one
Soldi R,Ghosh Halder T,Weston A,Thode T,Drenner K,Lewis R,Kaadige MR,Srivastava S,Daniel Ampanattu S,Rodriguez Del Villar R,Lang J,Vankayalapati H,Weissman B,Trent JM,Hendricks WPD,Sharma S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Sep 10 00:00:00 UTC 2020
In vitro-transcribed antigen receptor mRNA nanocarriers for transient expression in circulating T cells in vivo.
Nature communications
Parayath NN,Stephan SB,Koehne AL,Nelson PS,Stephan MT
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Nov 27 00:00:00 UTC 2020
α-Lactosylceramide Protects Against iNKT-Mediated Murine Airway Hyperreactivity and Liver Injury Through Competitive Inhibition of Cd1d Binding.
Frontiers in chemistry
Lai AC,Chi PY,Thio CL,Han YC,Kao HN,Hsieh HW,Gervay-Hague J,Chang YJ
14-0459 was used in Flow cytometry/Cell sorting to study how inhibiting iNKT cell activation with α-Lactosylceramide can reduce development of murine airway hyperreactivity and liver injury by competing for CD1d binding.
Thu Oct 01 00:00:00 UTC 2020
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
Side-by-side comparison of BH3-mimetics identifies MCL-1 as a key therapeutic target in AML.
Cell death & disease
Ewald L,Dittmann J,Vogler M,Fulda S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Wed Dec 04 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Mechanisms of virus dissemination in bone marrow of HIV-1-infected humanized BLT mice.
eLife
Ladinsky MS,Khamaikawin W,Jung Y,Lin S,Lam J,An DS,Bjorkman PJ,Kieffer C
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Mon Oct 28 00:00:00 UTC 2019
Si-ni-tang (a Chinese herbal formula) for improving immunofunction in sepsis: study protocol for a pilot randomized controlled trial.
Trials
Zeng R,Zheng Y,Fan R,Zhou G,Zhang Y,Mai S,Xie D,Weng Y,Du J,Han Y,Lai F
14-0459 was used in Flow cytometry/Cell sorting to study whether a Chinese herbal treatment, Si-ni-tang, can improve immunofunction in sepsis patients.
Wed Aug 28 00:00:00 UTC 2019
DKK2 imparts tumor immunity evasion through β-catenin-independent suppression of cytotoxic immune-cell activation.
Nature medicine
Xiao Q,Wu J,Wang WJ,Chen S,Zheng Y,Yu X,Meeth K,Sahraei M,Bothwell ALM,Chen L,Bosenberg M,Chen J,Sexl V,Sun L,Li L,Tang W,Wu D
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Mar 01 00:00:00 UTC 2018
Mast cells derived from human induced pluripotent stem cells are useful for allergen tests.
Allergology international : official journal of the Japanese Society of Allergology
Igarashi A,Ebihara Y,Kumagai T,Hirai H,Nagata K,Tsuji K
14-0459 was used in Flow cytometry/Cell sorting to introduce iMAT which may serve as a novel determining system for IgE/allergens in the clinical and research settings.
Sun Apr 01 00:00:00 UTC 2018
Single nucleotide variant profiles of viable single circulating tumour cells reveal CTC behaviours in breast cancer.
Oncology reports
Wang Y,Guo L,Feng L,Zhang W,Xiao T,Di X,Chen G,Zhang K
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue May 01 00:00:00 UTC 2018
Isolation of villous cytotrophoblasts from second trimester human placentas.
Placenta
Jankovic-Karasoulos T,McAninch D,McCullough D,Wilson RL,Bianco-Miotto T,Roberts CT
14-0459 was used in Immunohistochemistry-immunofluorescence to report a method for isolation and purification of primary villous cytotrophoblasts from 2nd trimester human placentas, that yields >99% trophoblast purity as shown by immunofluorescence.
Sat Dec 15 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
Fludarabine as a cost-effective adjuvant to enhance engraftment of human normal and malignant hematopoiesis in immunodeficient mice.
Scientific reports
Pievani A,Michelozzi IM,Rambaldi B,Granata V,Corsi A,Dazzi F,Biondi A,Serafini M
14-0459 was used in Flow cytometry/Cell sorting to present a novel and affordable model for engraftment of human normal and malignant hematopoiesis in immunodeficient mice.
Thu Jun 14 00:00:00 UTC 2018
Durable regression of Medulloblastoma after regional and intravenous delivery of anti-HER2 chimeric antigen receptor T cells.
Journal for immunotherapy of cancer
Nellan A,Rota C,Majzner R,Lester-McCully CM,Griesinger AM,Mulcahy Levy JM,Foreman NK,Warren KE,Lee DW
14-0459 was used in Flow cytometry/Cell sorting to evaluate the efficacy of anti-HER2 chimeric antigen receptor T cells for the treatment of paediatric medulloblastoma.
Mon Apr 30 00:00:00 UTC 2018
Expression of a Chimeric Antigen Receptor Specific for Donor HLA Class I Enhances the Potency of Human Regulatory T Cells in Preventing Human Skin Transplant Rejection.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Boardman DA,Philippeos C,Fruhwirth GO,Ibrahim MA,Hannen RF,Cooper D,Marelli-Berg FM,Watt FM,Lechler RI,Maher J,Smyth LA,Lombardi G
14-0459 was used in Immunohistochemistry to study the use of chimeric antigen receptor technology for the clinical refinement of regulatory T cell therapy for organ transplantation.
Sat Apr 01 00:00:00 UTC 2017
Evaluation of a simple method for storage of blood samples that enables isolation of circulating tumor cells 96 h after sample collection.
Journal of biological research (Thessalonike, Greece)
Apostolou P,Ntanovasilis DA,Papasotiriou I
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunofluorescence
Fri Dec 01 00:00:00 UTC 2017
Evaluation of a simple method for storage of blood samples that enables isolation of circulating tumor cells 96 h after sample collection.
Journal of biological research (Thessalonike, Greece)
Apostolou P,Ntanovasilis DA,Papasotiriou I
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunofluorescence
Fri Dec 01 00:00:00 UTC 2017
Evaluation of a simple method for storage of blood samples that enables isolation of circulating tumor cells 96 h after sample collection.
Journal of biological research (Thessalonike, Greece)
Apostolou P,Ntanovasilis DA,Papasotiriou I
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunofluorescence
Fri Dec 01 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Preferential accumulation of regulatory T cells with highly immunosuppressive characteristics in breast tumor microenvironment.
Oncotarget
Syed Khaja AS,Toor SM,El Salhat H,Faour I,Ul Haq N,Ali BR,Elkord E
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue May 16 00:00:00 UTC 2017
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Sep 12 00:00:00 UTC 2017
Deficient Adipogenesis of Scleroderma Patient and Healthy African American Monocytes.
Frontiers in pharmacology
Lee R,Reese C,Carmen-Lopez G,Perry B,Bonner M,Zemskova M,Wilson CL,Helke KL,Silver RM,Hoffman S,Tourkina E
14-0459 was used in Flow cytometry/Cell sorting to suggest that caveolin-1 and PPARγ work together to maintain a balance between the fibrogenic and adipogenic differentiation of monocytes.
Thu Oct 01 00:00:00 UTC 2020
Targeting methyltransferase PRMT5 eliminates leukemia stem cells in chronic myelogenous leukemia.
The Journal of clinical investigation
Jin Y,Zhou J,Xu F,Jin B,Cui L,Wang Y,Du X,Li J,Li P,Ren R,Pan J
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Mon Oct 03 00:00:00 UTC 2016
UTX inhibition as selective epigenetic therapy against TAL1-driven T-cell acute lymphoblastic leukemia.
Genes & development
Benyoucef A,Palii CG,Wang C,Porter CJ,Chu A,Dai F,Tremblay V,Rakopoulos P,Singh K,Huang S,Pflumio F,Hébert J,Couture JF,Perkins TJ,Ge K,Dilworth FJ,Brand M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
UTX inhibition as selective epigenetic therapy against TAL1-driven T-cell acute lymphoblastic leukemia.
Genes & development
Benyoucef A,Palii CG,Wang C,Porter CJ,Chu A,Dai F,Tremblay V,Rakopoulos P,Singh K,Huang S,Pflumio F,Hébert J,Couture JF,Perkins TJ,Ge K,Dilworth FJ,Brand M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
A 3D culture system enhances the ability of human bone marrow stromal cells to support the growth of limbal stem/progenitor cells.
Stem cell research
González S,Mei H,Nakatsu MN,Baclagon ER,Deng SX
Tue Mar 01 00:00:00 UTC 2016
Platelets confound the measurement of extracellular miRNA in archived plasma.
Scientific reports
Mitchell AJ,Gray WD,Hayek SS,Ko YA,Thomas S,Rooney K,Awad M,Roback JD,Quyyumi A,Searles CD
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Sep 13 00:00:00 UTC 2016
Circulating tumor cell detection in hepatocellular carcinoma based on karyoplasmic ratios using imaging flow cytometry.
Scientific reports
Liu Z,Guo W,Zhang D,Pang Y,Shi J,Wan S,Cheng K,Wang J,Cheng S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Dec 23 00:00:00 UTC 2016
Circulating tumor cell detection in hepatocellular carcinoma based on karyoplasmic ratios using imaging flow cytometry.
Scientific reports
Liu Z,Guo W,Zhang D,Pang Y,Shi J,Wan S,Cheng K,Wang J,Cheng S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Fri Dec 23 00:00:00 UTC 2016
Kinase-independent function of RIP1, critical for mature T-cell survival and proliferation.
Cell death & disease
Dowling JP,Cai Y,Bertin J,Gough PJ,Zhang J
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Sep 29 00:00:00 UTC 2016
Inactivation of nuclear GSK3β by Ser(389) phosphorylation promotes lymphocyte fitness during DNA double-strand break response.
Nature communications
Thornton TM,Delgado P,Chen L,Salas B,Krementsov D,Fernandez M,Vernia S,Davis RJ,Heimann R,Teuscher C,Krangel MS,Ramiro AR,Rincón M
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Western Blot
Fri Jan 29 00:00:00 UTC 2016
Multivalent Chromatin Engagement and Inter-domain Crosstalk Regulate MORC3 ATPase.
Cell reports
Andrews FH,Tong Q,Sullivan KD,Cornett EM,Zhang Y,Ali M,Ahn J,Pandey A,Guo AH,Strahl BD,Costello JC,Espinosa JM,Rothbart SB,Kutateladze TG
14-0459 was used in Flow cytometry/Cell sorting to demonstrate the interaction between MORC3 and methylated histone H3K4 tails and implicate it''s role in human disease.
Tue Sep 20 00:00:00 UTC 2016
A novel function of RIP1 in postnatal development and immune homeostasis by protecting against RIP3-dependent necroptosis and FADD-mediated apoptosis.
Frontiers in cell and developmental biology
Dowling JP,Nair A,Zhang J
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Western Blot
Fri Mar 13 00:00:00 UTC 2015
Activation of the Wnt/planar cell polarity pathway is required for pericyte recruitment during pulmonary angiogenesis.
The American journal of pathology
Yuan K,Orcholski ME,Panaroni C,Shuffle EM,Huang NF,Jiang X,Tian W,Vladar EK,Wang L,Nicolls MR,Wu JY,de Jesus Perez VA
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Retinoic acid regulates hematopoietic development from human pluripotent stem cells.
Stem cell reports
Rönn RE,Guibentif C,Moraghebi R,Chaves P,Saxena S,Garcia B,Woods NB
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Feb 10 00:00:00 UTC 2015
Reprogramming human endothelial cells to haematopoietic cells requires vascular induction.
Nature
Sandler VM,Lis R,Liu Y,Kedem A,James D,Elemento O,Butler JM,Scandura JM,Rafii S
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Jul 17 00:00:00 UTC 2014
Prognostic significance of CD44 variant 2 upregulation in colorectal cancer.
British journal of cancer
Ozawa M,Ichikawa Y,Zheng YW,Oshima T,Miyata H,Nakazawa K,Guan HB,Shiozawa M,Akaike M,Watanabe K,Ota M,Fujii S,Kunisaki C,Ishikawa T,Tanaka K,Akiyama H,Endo I,Taniguchi H
14-0459 was used in Flow cytometry/Cell sorting to investigate the clinical significance of CD133 and CD44 as putative cancer stem cell markers.
Tue Jul 15 00:00:00 UTC 2014
Caveolin-1 regulates chemokine receptor 5-mediated contribution of bone marrow-derived cells to dermal fibrosis.
Frontiers in pharmacology
Lee R,Perry B,Heywood J,Reese C,Bonner M,Hatfield CM,Silver RM,Visconti RP,Hoffman S,Tourkina E
Thu Jun 26 00:00:00 UTC 2014
Caveolin-1 regulates chemokine receptor 5-mediated contribution of bone marrow-derived cells to dermal fibrosis.
Frontiers in pharmacology
Lee R,Perry B,Heywood J,Reese C,Bonner M,Hatfield CM,Silver RM,Visconti RP,Hoffman S,Tourkina E
Thu Jun 26 00:00:00 UTC 2014
Caveolin-1 regulates chemokine receptor 5-mediated contribution of bone marrow-derived cells to dermal fibrosis.
Frontiers in pharmacology
Lee R,Perry B,Heywood J,Reese C,Bonner M,Hatfield CM,Silver RM,Visconti RP,Hoffman S,Tourkina E
Thu Jun 26 00:00:00 UTC 2014
A Small Molecule β2 Integrin Agonist Improves Chronic Kidney Allograft Survival by Reducing Leukocyte Recruitment and Accompanying Vasculopathy.
Frontiers in medicine
Khan SQ,Guo L,Cimbaluk DJ,Elshabrawy H,Faridi MH,Jolly M,George JF,Agarwal A,Gupta V
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunofluorescence
Fri Jan 16 00:00:00 UTC 2015
A Small Molecule β2 Integrin Agonist Improves Chronic Kidney Allograft Survival by Reducing Leukocyte Recruitment and Accompanying Vasculopathy.
Frontiers in medicine
Khan SQ,Guo L,Cimbaluk DJ,Elshabrawy H,Faridi MH,Jolly M,George JF,Agarwal A,Gupta V
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Immunofluorescence
Fri Jan 16 00:00:00 UTC 2015
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
Monocytes from chronic HBV patients react in vitro to HBsAg and TLR by producing cytokines irrespective of stage of disease.
PloS one
Boltjes A,Groothuismink ZM,van Oord GW,Janssen HL,Woltman AM,Boonstra A
14-0459 was used in Flow cytometry/Cell sorting to investigate the in vitro and in vivo effects of hepatitis B virus surface antigen on monocytes from chronic HBV patients.
Tue Jan 06 00:00:00 UTC 2015
Human fetal liver cells for regulated ex vivo erythropoietin gene therapy.
Molecular therapy. Methods & clinical development
El Filali E,Duijst S,Hiralall JK,Legrand N,van Gulik T,Hoekstra R,Seppen J
14-0459 was used in Immunohistochemistry to investigate the utility of human fetal liver cells for therapeutic protein delivery.
Wed May 27 00:00:00 UTC 2015
Human fetal liver cells for regulated ex vivo erythropoietin gene therapy.
Molecular therapy. Methods & clinical development
El Filali E,Duijst S,Hiralall JK,Legrand N,van Gulik T,Hoekstra R,Seppen J
14-0459 was used in Immunohistochemistry to investigate the utility of human fetal liver cells for therapeutic protein delivery.
Wed May 27 00:00:00 UTC 2015
Clinical grade manufacturing of human alloantigen-reactive regulatory T cells for use in transplantation.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons
Putnam AL,Safinia N,Medvec A,Laszkowska M,Wray M,Mintz MA,Trotta E,Szot GL,Liu W,Lares A,Lee K,Laing A,Lechler RI,Riley JL,Bluestone JA,Lombardi G,Tang Q
14-0459 was used in Immunofluorescence in immunohistochemistry on frozen tissues to develop a manufacturing process to expand human alloantigen-reactive regulatory T cells in short-term cultures using good manufacturing practice-compliant reagents.
Fri Nov 01 00:00:00 UTC 2013
Milky spots promote ovarian cancer metastatic colonization of peritoneal adipose in experimental models.
The American journal of pathology
Clark R,Krishnan V,Schoof M,Rodriguez I,Theriault B,Chekmareva M,Rinker-Schaeffer C
14-0459 was used in Immunohistochemistry to support a two-step model in which both milky spots and adipose have specific roles in colonization of the omentum by ovarian cancer cells.
Thu Aug 01 00:00:00 UTC 2013
Microvesicles derived from human umbilical cord Wharton's jelly mesenchymal stem cells attenuate bladder tumor cell growth in vitro and in vivo.
PloS one
Wu S,Ju GQ,Du T,Zhu YJ,Liu GH
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Tue Oct 22 00:00:00 UTC 2013
Human regulatory T cells with alloantigen specificity are more potent inhibitors of alloimmune skin graft damage than polyclonal regulatory T cells.
Science translational medicine
Sagoo P,Ali N,Garg G,Nestle FO,Lechler RI,Lombardi G
14-0459 was used in Immunohistochemistry to develop a protocol for the selective production of alloantigen-specific regulatory T cells, and compare their efficacy in inhibiting alloimmune damage with that of polyclonal regulatory T cells.
Wed May 18 00:00:00 UTC 2011
Yield and characterization of subcutaneous human adipose-derived stem cells by flow cytometric and adipogenic mRNA analyzes.
Cytotherapy
Yu G,Wu X,Dietrich MA,Polk P,Scott LK,Ptitsyn AA,Gimble JM
Published figure using CD45 monoclonal antibody (Product # 14-0459-82) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2010
Lung dendritic cell expression of maturation molecules increases with worsening chronic obstructive pulmonary disease.
American journal of respiratory and critical care medicine
Freeman CM,Martinez FJ,Han MK,Ames TM,Chensue SW,Todt JC,Arenberg DA,Meldrum CA,Getty C,McCloskey L,Curtis JL
14-0459 was used in Flow cytometry/Cell sorting to determine the expression of maturation molecules by individual DC subsets in relationship to COPD stage.
Tue Dec 15 00:00:00 UTC 2009
C-terminal repeats of Clostridium difficile toxin A induce production of chemokine and adhesion molecules in endothelial cells and promote migration of leukocytes.
Infection and immunity
Yeh CY,Lin CN,Chang CF,Lin CH,Lien HT,Chen JY,Chia JS
14-0459 was used in Flow cytometry/Cell sorting to investigate how Clostridium difficile toxin A may bind to multiple carbohydrate motifs to exert its biological activity on human endothelial cells.
Sat Mar 01 00:00:00 UTC 2008
A new protocol for multiple inhalation of IFN-gamma successfully treats MDR-TB: a case study.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease
Grahmann PR,Braun RK
14045982 was used in flow cytometry to demonstrate that aerosolized interferon-gamma results in the transient conversion of sputum smears in multidrug-resistant pulmonary tuberculosis
Sun Jun 01 00:00:00 UTC 2008