Product References
VentX promotes tumor specific immunity and efficacy of immune checkpoint inhibitors.
iScience
Le Y,Gao H,Le J,Hornick JL,Bleday R,Wee J,Zhu Z
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Fri Jan 19 00:00:00 UTC 2024
Activation of the JNK/COX-2/HIF-1α axis promotes M1 macrophage via glycolytic shift in HIV-1 infection.
Life science alliance
Zhang J,Yuan Z,Li X,Wang F,Wei X,Kang Y,Mo C,Jiang J,Liang H,Ye L
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Inhibition of inflammation and infiltration of M2 macrophages in NSCLC through the ATF3/CSF1 axis: Role of miR-27a-3p.
International journal of experimental pathology
Zhou B,Xu Y,Xu L,Kong Y,Li K,Chen B,Li J
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Targeting Glioblastoma-Associated Macrophages for Photodynamic Therapy Using AGuIX®-Design Nanoparticles.
Pharmaceutics
Lerouge L,Gries M,Chateau A,Daouk J,Lux F,Rocchi P,Cedervall J,Olsson AK,Tillement O,Frochot C,Acherar S,Thomas N,Barberi-Heyob M
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry
Mon Mar 20 00:00:00 UTC 2023
Complete Freund's Adjuvant Induces a Fibroblast-like Synoviocytes (FLS) Metabolic and Migratory Phenotype in Resident Fibroblasts of the Inoculated Footpad at the Earliest Stage of Adjuvant-Induced Arthritis.
Cells
González-Chávez SA,Chaparro-Barrera E,Alvarado-Jáquez MF,Cuevas-Martínez R,Ochoa-Albíztegui RE,Pacheco-Tena C
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry
Wed Mar 08 00:00:00 UTC 2023
LINC00240/miR-155 axis regulates function of trophoblasts and M2 macrophage polarization via modulating oxidative stress-induced pyroptosis in preeclampsia.
Molecular medicine (Cambridge, Mass.)
Wu HY,Liu K,Zhang JL
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Sat Sep 24 00:00:00 UTC 2022
PTX3 mediates the infiltration, migration, and inflammation-resolving-polarization of macrophages in glioblastoma.
CNS neuroscience & therapeutics
Zhang H,Wang Y,Zhao Y,Liu T,Wang Z,Zhang N,Dai Z,Wu W,Cao H,Feng S,Zhang L,Cheng Q,Liu Z
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
Divide and Conquer: Isolating Cell Populations to Investigate How Breast Cancer Risk Factors Alter the Breast Microenvironment.
Methods in molecular biology (Clifton, N.J.)
Arendt LM
14-0689-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) to investigate how risk factors impact distinct cell populations within breast tissue.
Mon Feb 21 00:00:00 UTC 2022
Different Induction of PD-L1 (CD274) and PD-1 (CD279) Expression in THP-1-Differentiated Types 1 and 2 Macrophages.
Journal of inflammation research
Lai CY,Tseng PC,Chen CL,Satria RD,Wang YT,Lin CF
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Wed Apr 27 00:00:00 UTC 2022
Decreased ferroportin in hepatocytes promotes macrophages polarize towards an M2-like phenotype and liver fibrosis.
Scientific reports
Cai C,Zeng D,Gao Q,Ma L,Zeng B,Zhou Y,Wang H
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry (Paraffin)
Mon Jun 28 00:00:00 UTC 2021
Antitumor properties of triptolide: phenotype regulation of macrophage differentiation.
Cancer biology & therapy
Li H,Li L,Mei H,Pan G,Wang X,Huang X,Wang T,Jiang Z,Zhang L,Sun L
14-0689-82 was used in Flow Cytometry to determine whether triptolide has any effect on the activation of tumor-associated macrophages and the production of tumor-promoting mediators.
Mon Mar 29 00:00:00 UTC 2021
The role of CD68+ macrophage in classical Hodgkin lymphoma patients from Egypt.
Diagnostic pathology
Mohamed O,El Bastawisy A,Allahlobi N,Abdellateif MS,Zekri ARN,Shaarawy S,Korany Z,Mohanad M,Bahnassy AA
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Tue Feb 04 00:00:00 UTC 2020
The role of CD68+ macrophage in classical Hodgkin lymphoma patients from Egypt.
Diagnostic pathology
Mohamed O,El Bastawisy A,Allahlobi N,Abdellateif MS,Zekri ARN,Shaarawy S,Korany Z,Mohanad M,Bahnassy AA
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Tue Feb 04 00:00:00 UTC 2020
Nanotopographical Coatings Induce an Early Phenotype-Specific Response of Primary Material-Resident M1 and M2 Macrophages.
Materials (Basel, Switzerland)
Schmitz T,Jannasch M,Weigel T,Moseke C,Gbureck U,Groll J,Walles H,Hansmann J
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunomicroscopy
Wed Mar 04 00:00:00 UTC 2020
An altered response in macrophage phenotype following damage in aged human skeletal muscle: implications for skeletal muscle repair.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Sorensen JR,Kaluhiokalani JP,Hafen PS,Deyhle MR,Parcell AC,Hyldahl RD
14-0689 was used in Immunohistochemistry to test the hypothesis that macrophage polarisation is altered in old compared to young skeletal muscle, possibly contributing to the poor satellite cell response observed in older muscle tissue.
Sun Sep 01 00:00:00 UTC 2019
Identification of TNFR2 and IL-33 as therapeutic targets in localized fibrosis.
Science advances
Izadi D,Layton TB,Williams L,McCann F,Cabrita M,Espirito Santo AI,Xie W,Fritzsche M,Colin-York H,Feldmann M,Midwood KS,Nanchahal J
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Sun Dec 01 00:00:00 UTC 2019
Macrophagic CD146 promotes foam cell formation and retention during atherosclerosis.
Cell research
Luo Y,Duan H,Qian Y,Feng L,Wu Z,Wang F,Feng J,Yang D,Qin Z,Yan X
14-0689 was used in Immunofluorescence to identify CD146 as a retention signal that traps macrophages within the artery wall, and a promising therapeutic target in atherosclerosis treatment.
Wed Mar 01 00:00:00 UTC 2017
Immortalization of primary microglia: a new platform to study HIV regulation in the central nervous system.
Journal of neurovirology
Garcia-Mesa Y,Jay TR,Checkley MA,Luttge B,Dobrowolski C,Valadkhan S,Landreth GE,Karn J,Alvarez-Carbonell D
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
Suppression of the inflammatory response by disease-inducible interleukin-10 gene therapy in a three-dimensional micromass model of the human synovial membrane.
Arthritis research & therapy
Broeren MG,de Vries M,Bennink MB,Arntz OJ,van Lent PL,van der Kraan PM,van den Berg WB,van den Hoogen FH,Koenders MI,van de Loo FA
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry
Fri Aug 12 00:00:00 UTC 2016
MicroRNA-720 suppresses M2 macrophage polarization by targeting GATA3.
Bioscience reports
Zhong Y,Yi C
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunocytochemistry
Fri Dec 02 00:00:00 UTC 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunocytochemistry
Fri Dec 02 00:00:00 UTC 2016
Expression of Lectin-Like Transcript 1, the Ligand for CD161, in Rheumatoid Arthritis.
PloS one
Chalan P,Bijzet J,Huitema MG,Kroesen BJ,Brouwer E,Boots AM
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry
Mon Apr 11 00:00:00 UTC 2016
Doxycycline inhibits polarization of macrophages to the proangiogenic M2-type and subsequent neovascularization.
The Journal of biological chemistry
He L,Marneros AG
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Fri Mar 21 00:00:00 UTC 2014
Doxycycline inhibits polarization of macrophages to the proangiogenic M2-type and subsequent neovascularization.
The Journal of biological chemistry
He L,Marneros AG
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Flow Cytometry
Fri Mar 21 00:00:00 UTC 2014
Glioma grade is associated with the accumulation and activity of cells bearing M2 monocyte markers.
Clinical cancer research : an official journal of the American Association for Cancer Research
Prosniak M,Harshyne LA,Andrews DW,Kenyon LC,Bedelbaeva K,Apanasovich TV,Heber-Katz E,Curtis MT,Cotzia P,Hooper DC
14-0689 was used in Flow cytometry/Cell sorting to show that the content of CD163(+) cells distinguishes grade III astrocytoma subsets with different prognosis.
Mon Jul 15 00:00:00 UTC 2013
Hepatocellular proliferation correlates with inflammatory cell and cytokine changes in a murine model of nonalchoholic fatty liver disease.
PloS one
Vansaun MN,Mendonsa AM,Lee Gorden D
Published figure using CD68 monoclonal antibody (Product # 14-0689-82) in Immunohistochemistry
Sun Jun 15 00:00:00 UTC 2014
A consensus surface activation marker signature is partially dependent on human immunodeficiency virus type 1 Nef expression within productively infected macrophages.
Retrovirology
Babu R,Brown A
14-0689 was used in Flow cytometry/Cell sorting to determine whether macrophage surface marker phenotype in non-polarised cultures is altered by HIV infection independently of host genetic background.
Mon Dec 16 00:00:00 UTC 2013