Product References
Proteolytic activation of angiomotin by DDI2 promotes angiogenesis.
The EMBO journal
Wang Y,Zhu Y,Wang Y,Chang Y,Geng F,Ma M,Gu Y,Yu A,Zhu R,Yu P,Sha Z,Qi S,Li J,Zhao W,Pan W,Zhang R,Yu FX
14-1441-82 was used in Immunohistochemistry to report that AMOT undergoes cleavage in the presence of lysophosphatidic acid (LPA), a lipid growth factor also involved in angiogenesis.
Tue Aug 01 00:00:00 UTC 2023
Flow pattern-dependent mitochondrial dynamics regulates the metabolic profile and inflammatory state of endothelial cells.
JCI insight
Hong SG,Shin J,Choi SY,Powers JC,Meister BM,Sayoc J,Son JS,Tierney R,Recchia FA,Brown MD,Yang X,Park JY
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunohistochemistry
Thu Sep 22 00:00:00 UTC 2022
Robo4 is constitutively shed by ADAMs from endothelial cells and the shed Robo4 functions to inhibit Slit3-induced angiogenesis.
Scientific reports
Xiao W,Pinilla-Baquero A,Faulkner J,Song X,Prabhakar P,Qiu H,Moremen KW,Ludwig A,Dempsey PJ,Azadi P,Wang L
14-1441-82 was used in Western Blot to delineate ADAM10 and ADAM17 are Robo4 sheddases, and ectodomain shedding, including negative regulation by its ligand Slit3, represents a novel control mechanism of Robo4 signaling in angiogenesis.
Mon Mar 14 00:00:00 UTC 2022
Altered Mitochondrial Opa1-Related Fusion in Mouse Promotes Endothelial Cell Dysfunction and Atherosclerosis.
Antioxidants (Basel, Switzerland)
Chehaitly A,Guihot AL,Proux C,Grimaud L,Aurrière J,Legouriellec B,Rivron J,Vessieres E,Tétaud C,Zorzano A,Procaccio V,Joubaud F,Reynier P,Lenaers G,Loufrani L,Henrion D
14-1441-82 was used in Immunohistochemistry-immunofluorescence to conclude that reduction in mitochondrial fusion in mouse ECs altered the dilator response to shear stress due to excessive superoxide production and induced greater atherosclerosis development.
Sat May 28 00:00:00 UTC 2022
Mfsd2b and Spns2 are essential for maintenance of blood vessels during development and in anaphylactic shock.
Cell reports
Le TNU,Nguyen TQ,Kalailingam P,Nguyen YTK,Sukumar VK,Tan CKH,Tukijan F,Couty L,Hasan Z,Del Gaudio I,Wenk MR,Cazenave-Gassiot A,Camerer E,Nguyen LN
14-1441-82 was used in Immunohistochemistry to show that S1P export from Mfsd2b and Spns2 is essential for developing and mature vasculature.
Tue Aug 16 00:00:00 UTC 2022
Monomeric C-reactive protein via endothelial CD31 for neurovascular inflammation in an ApoE genotype-dependent pattern: A risk factor for Alzheimer's disease?
Aging cell
Zhang Z,Na H,Gan Q,Tao Q,Alekseyev Y,Hu J,Yan Z,Yang JB,Tian H,Zhu S,Li Q,Rajab IM,Blusztajn JK,Wolozin B,Emili A,Zhang X,Stein T,Potempa LA,Qiu WQ
14-1441-82 was used in Immunohistochemistry to demonstrate that the interactions of endothelial CD31 with monomeric C-reactive protein (mCRP) versus ApoE were linked with shortened neurovasculature for Alzheimer's disease (AD) pathology and cognition.
Mon Nov 01 00:00:00 UTC 2021
Monomeric C-reactive protein via endothelial CD31 for neurovascular inflammation in an ApoE genotype-dependent pattern: A risk factor for Alzheimer's disease?
Aging cell
Zhang Z,Na H,Gan Q,Tao Q,Alekseyev Y,Hu J,Yan Z,Yang JB,Tian H,Zhu S,Li Q,Rajab IM,Blusztajn JK,Wolozin B,Emili A,Zhang X,Stein T,Potempa LA,Qiu WQ
14-1441-82 was used in Immunohistochemistry to demonstrate that the interactions of endothelial CD31 with monomeric C-reactive protein (mCRP) versus ApoE were linked with shortened neurovasculature for Alzheimer's disease (AD) pathology and cognition.
Mon Nov 01 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Western Blot
Tue May 11 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Western Blot
Tue May 11 00:00:00 UTC 2021
Knockout RAGE alleviates cardiac fibrosis through repressing endothelial-to-mesenchymal transition (EndMT) mediated by autophagy.
Cell death & disease
Zhang L,He J,Wang J,Liu J,Chen Z,Deng B,Wei L,Wu H,Liang B,Li H,Huang Y,Lu L,Yang Z,Xian S,Wang L
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Western Blot
Tue May 11 00:00:00 UTC 2021
VE-cadherin endocytosis controls vascular integrity and patterning during development.
The Journal of cell biology
Grimsley-Myers CM,Isaacson RH,Cadwell CM,Campos J,Hernandes MS,Myers KR,Seo T,Giang W,Griendling KK,Kowalczyk AP
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunohistochemistry
Mon May 04 00:00:00 UTC 2020
VE-cadherin endocytosis controls vascular integrity and patterning during development.
The Journal of cell biology
Grimsley-Myers CM,Isaacson RH,Cadwell CM,Campos J,Hernandes MS,Myers KR,Seo T,Giang W,Griendling KK,Kowalczyk AP
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunohistochemistry
Mon May 04 00:00:00 UTC 2020
VE-cadherin endocytosis controls vascular integrity and patterning during development.
The Journal of cell biology
Grimsley-Myers CM,Isaacson RH,Cadwell CM,Campos J,Hernandes MS,Myers KR,Seo T,Giang W,Griendling KK,Kowalczyk AP
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunohistochemistry
Mon May 04 00:00:00 UTC 2020
Interleukin 35 Delays Hindlimb Ischemia-Induced Angiogenesis Through Regulating ROS-Extracellular Matrix but Spares Later Regenerative Angiogenesis.
Frontiers in immunology
Fu H,Sun Y,Shao Y,Saredy J,Cueto R,Liu L,Drummer C,Johnson C,Xu K,Lu Y,Li X,Meng S,Xue ER,Tan J,Jhala NC,Yu D,Zhou Y,Bayless KJ,Yu J,Rogers TJ,Hu W,Snyder NW,Sun J,Qin X,Jiang X,Wang H,Yang X
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Fri Jul 09 00:00:00 UTC 2021
Possible application of muscle specific conditional mouse-derived induced pluripotent stem cells for muscle research.
Biochemistry and biophysics reports
Hosoyama T
14-1441-82 was used in Immunocytochemistry to examine whether homologous recombination was induced in mCM-iPSC-derived myotubes by 4-hydroxytamoxifen (4OH-TAM) administration.
Sun Mar 01 00:00:00 UTC 2020
Cellular cartography of the organ of Corti based on optical tissue clearing and machine learning.
eLife
Urata S,Iida T,Yamamoto M,Mizushima Y,Fujimoto C,Matsumoto Y,Yamasoba T,Okabe S
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunohistochemistry
Fri Jan 18 00:00:00 UTC 2019
CNS lymphatic drainage and neuroinflammation are regulated by meningeal lymphatic vasculature.
Nature neuroscience
Louveau A,Herz J,Alme MN,Salvador AF,Dong MQ,Viar KE,Herod SG,Knopp J,Setliff JC,Lupi AL,Da Mesquita S,Frost EL,Gaultier A,Harris TH,Cao R,Hu S,Lukens JR,Smirnov I,Overall CC,Oliver G,Kipnis J
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunocytochemistry
Mon Oct 01 00:00:00 UTC 2018
Role of Notch1 in the arterial specification and angiogenic potential of mouse embryonic stem cell-derived endothelial cells.
Stem cell research & therapy
Park JK,Lee TW,Do EK,Moon HJ,Kim JH
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Wed Jul 18 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Tue Mar 20 00:00:00 UTC 2018
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunocytochemistry
Thu May 04 00:00:00 UTC 2017
Tumour ischaemia by interferon-γ resembles physiological blood vessel regression.
Nature
Kammertoens T,Friese C,Arina A,Idel C,Briesemeister D,Rothe M,Ivanov A,Szymborska A,Patone G,Kunz S,Sommermeyer D,Engels B,Leisegang M,Textor A,Fehling HJ,Fruttiger M,Lohoff M,Herrmann A,Yu H,Weichselbaum R,Uckert W,Hübner N,Gerhardt H,Beule D,Schreiber H,Blankenstein T
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunocytochemistry
Thu May 04 00:00:00 UTC 2017
VEGFR3 Modulates Vascular Permeability by Controlling VEGF/VEGFR2 Signaling.
Circulation research
Heinolainen K,Karaman S,D'Amico G,Tammela T,Sormunen R,Eklund L,Alitalo K,Zarkada G
14-1441 was used in Immunofluorescence to examine vascular endothelial growth factor pathway activity in quiescent and angiogenic blood vascular endothelial cells.
Fri Apr 28 00:00:00 UTC 2017
The VE-cadherin cytoplasmic domain undergoes proteolytic processing during endocytosis.
Molecular biology of the cell
Su W,Kowalczyk AP
14144182 was used in western blot to study the trafficking of VE-cadherin after endocytosis
Sun Jan 01 00:00:00 UTC 2017
Activation of the TGFβ pathway impairs endothelial to haematopoietic transition.
Scientific reports
Vargel Ö,Zhang Y,Kosim K,Ganter K,Foehr S,Mardenborough Y,Shvartsman M,Enright AJ,Krijgsveld J,Lancrin C
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Fri Feb 19 00:00:00 UTC 2016
Suppression of ischemia in arterial occlusive disease by JNK-promoted native collateral artery development.
eLife
Ramo K,Sugamura K,Craige S,Keaney JF,Davis RJ
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Western Blot
Tue Aug 09 00:00:00 UTC 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Thu Jun 02 00:00:00 UTC 2016
Structural and functional features of central nervous system lymphatic vessels.
Nature
Louveau A,Smirnov I,Keyes TJ,Eccles JD,Rouhani SJ,Peske JD,Derecki NC,Castle D,Mandell JW,Lee KS,Harris TH,Kipnis J
14-1441 was used in Immunohistochemistry-wholemount to investigate the mechanisms governing the entrance and exit of immune cells from the CNS.
Thu Jul 16 00:00:00 UTC 2015
Endothelial destabilization by angiopoietin-2 via integrin β1 activation.
Nature communications
Hakanpaa L,Sipila T,Leppanen VM,Gautam P,Nurmi H,Jacquemet G,Eklund L,Ivaska J,Alitalo K,Saharinen P
14-1441 was used in Immunocytochemistry-immunoflourescence to elucidate the Angiopoietin-2-dependent mechanisms of vascular destabilisation.
Fri Jan 30 00:00:00 UTC 2015
Endothelial destabilization by angiopoietin-2 via integrin β1 activation.
Nature communications
Hakanpaa L,Sipila T,Leppanen VM,Gautam P,Nurmi H,Jacquemet G,Eklund L,Ivaska J,Alitalo K,Saharinen P
14-1441 was used in Immunocytochemistry-immunoflourescence to elucidate the Angiopoietin-2-dependent mechanisms of vascular destabilisation.
Fri Jan 30 00:00:00 UTC 2015
Endothelial fate mapping in mice with pulmonary hypertension.
Circulation
Qiao L,Nishimura T,Shi L,Sessions D,Thrasher A,Trudell JR,Berry GJ,Pearl RG,Kao PN
14-1441 was used in Immunofluorescence to demonstrate that a fraction of neointimal cells in pulmonary hypertension exhibit an endothelial lineage of origin while actively expressing smooth muscle genes.
Tue Feb 11 00:00:00 UTC 2014
Angiopoietin 2 regulates the transformation and integrity of lymphatic endothelial cell junctions.
Genes & development
Zheng W,Nurmi H,Appak S,Sabine A,Bovay E,Korhonen EA,Orsenigo F,Lohela M,D'Amico G,Holopainen T,Leow CC,Dejana E,Petrova TV,Augustin HG,Alitalo K
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunofluorescence
Tue Jul 15 00:00:00 UTC 2014
Sox7 is regulated by ETV2 during cardiovascular development.
Stem cells and development
Behrens AN,Zierold C,Shi X,Ren Y,Koyano-Nakagawa N,Garry DJ,Martin CM
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Mon Sep 01 00:00:00 UTC 2014
The expression of Sox17 identifies and regulates haemogenic endothelium.
Nature cell biology
Clarke RL,Yzaguirre AD,Yashiro-Ohtani Y,Bondue A,Blanpain C,Pear WS,Speck NA,Keller G
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Flow Cytometry
Wed May 01 00:00:00 UTC 2013
Sphingosine-1-phosphate receptor 2 protects against anaphylactic shock through suppression of endothelial nitric oxide synthase in mice.
The Journal of allergy and clinical immunology
Cui H,Okamoto Y,Yoshioka K,Du W,Takuwa N,Zhang W,Asano M,Shibamoto T,Takuwa Y
14-1441 was used in Immunofluorescence to suggest the potential usefulness of Sphingosine-1-phosphate receptor 2 agonists as novel therapeutic agents for anaphylaxis.
Fri Nov 01 00:00:00 UTC 2013
HIF-1α activation results in actin cytoskeleton reorganization and modulation of Rac-1 signaling in endothelial cells.
Cell communication and signaling : CCS
Weidemann A,Breyer J,Rehm M,Eckardt KU,Daniel C,Cicha I,Giehl K,Goppelt-Struebe M
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunofluorescence
Mon Oct 21 00:00:00 UTC 2013
HIF-1α activation results in actin cytoskeleton reorganization and modulation of Rac-1 signaling in endothelial cells.
Cell communication and signaling : CCS
Weidemann A,Breyer J,Rehm M,Eckardt KU,Daniel C,Cicha I,Giehl K,Goppelt-Struebe M
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunofluorescence
Mon Oct 21 00:00:00 UTC 2013
HIF-1α activation results in actin cytoskeleton reorganization and modulation of Rac-1 signaling in endothelial cells.
Cell communication and signaling : CCS
Weidemann A,Breyer J,Rehm M,Eckardt KU,Daniel C,Cicha I,Giehl K,Goppelt-Struebe M
Published figure using CD144 (VE-cadherin) monoclonal antibody (Product # 14-1441-82) in Immunofluorescence
Mon Oct 21 00:00:00 UTC 2013
Discontinuous LYVE-1 expression in corneal limbal lymphatics: dual function as microvalves and immunological hot spots.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Nakao S,Zandi S,Faez S,Kohno R,Hafezi-Moghadam A
14-1441 was used in Immunofluorescence to investigate how LYVE-1(-) lymphatic endothelium could serve as microvalves and immunological hot spots.
Wed Feb 01 00:00:00 UTC 2012
Snail promotes the cell-autonomous generation of Flk1(+) endothelial cells through the repression of the microRNA-200 family.
Stem cells and development
Gill JG,Langer EM,Lindsley RC,Cai M,Murphy TL,Murphy KM
14-1441 was used in Flow cytometry/Cell sorting to assess the role of snail to promote the cell-autonomous induction of Flk1(+) endothelial cells in mouse embryonic stem cells.
Fri Jan 20 00:00:00 UTC 2012
Targeting Robo4-dependent Slit signaling to survive the cytokine storm in sepsis and influenza.
Science translational medicine
London NR,Zhu W,Bozza FA,Smith MC,Greif DM,Sorensen LK,Chen L,Kaminoh Y,Chan AC,Passi SF,Day CW,Barnard DL,Zimmerman GA,Krasnow MA,Li DY
14-1441 was used in Blocking experiments to explore strengthening the vascular barrier as a method for combating septic shock in multiple animal models of infection.
Wed Mar 17 00:00:00 UTC 2010
Three-dimensional cartography of hematopoietic clusters in the vasculature of whole mouse embryos.
Development (Cambridge, England)
Yokomizo T,Dzierzak E
14-1441 was used in Immunohistochemistry to devise a method to make transparent whole mouse embryos for the observation of hematopoietic clusters.
Mon Nov 01 00:00:00 UTC 2010
Endothelial-myofibroblast transition contributes to the early development of diabetic renal interstitial fibrosis in streptozotocin-induced diabetic mice.
The American journal of pathology
Li J,Qu X,Bertram JF
Thu Oct 01 00:00:00 UTC 2009