Invitrogen antibodies—maximize lab budgets with advanced validation & coverage
Shop antibodies
    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • VEGF Receptor 3 Antibodies
            Invitrogen
            VEGF Receptor 3 Monoclonal Antibody (AFL4), eBioscience™
            Advanced Verification
            11 Published Figures
            23 References
            View all (35) VEGF Receptor 3 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 14-5988-85
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            Price (USD)
            286.45
            Special offer
            Online exclusive
            337.00
            Save 50.55 (15%)
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers
            Cite VEGF Receptor 3 Monoclonal Antibody (AFL4), eBioscience™
            • Published Figures (11)
            • Advanced Verification (1)
            Application
            VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
            Group 53 Created with Sketch.
            FIGURE: 1 / 12
            VEGF Receptor 3 Antibody (14-5988-85) in IHC
            Life science alliance 2022 - Figure S5. Increased Vegfr3 expression induced by Tbx1 Cre activation of TgVegfr3 is revealed by qRT-PCR but not by immunofluorescence. (A, B, C, D, E, F, G, H, I, J, K, L) Representative coronal brain sections of Tbx1 Cre/ + (A, B, C, D, E, F) and TgVegfr3; Tbx1 Cre/ + (G, H, I, J, K, L) embryos (E18.5) stained for VEGFR3 (red), VEGFR2 (green), and GFP ( Tbx1 cell fate, white). The cartoon shows the positi... View More
            View Product
            VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
            VEGF Receptor 3 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
            VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
            VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
            VEGF Receptor 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
            VEGF Receptor 3 Antibody in Immunohistochemistry (IHC)
            VEGF Receptor 3 Antibody in Western Blot (WB)
            VEGF Receptor 3 Antibody in Western Blot (WB)
            VEGF Receptor 3 Antibody in Western Blot (WB)
            VEGF Receptor 3 Antibody in Flow Cytometry (Flow)
            VEGF Receptor 3 Antibody in Flow Cytometry (Flow)
            VEGF Receptor 3 Antibody
            Product Details
            14-5988-85
            Applications
            Tested Dilution
            Publications
            Western Blot (WB)
            Assay-Dependent
            View 7 publications 7 publications
            Immunohistochemistry (IHC)
            -
            View 9 publications 9 publications
            Immunohistochemistry (Paraffin) (IHC (P))
            -
            View 1 publication 1 publication
            Immunohistochemistry (Frozen) (IHC (F))
            Assay-Dependent
            View 1 publication 1 publication
            Immunocytochemistry (ICC/IF)
            -
            View 1 publication 1 publication
            Flow Cytometry (Flow)
            Assay-Dependent
            View 3 publications 3 publications
            Immunoprecipitation (IP)
            Assay-Dependent
            View 1 publication 1 publication
            Neutralization (Neu)
            Assay-Dependent
            -
            Functional Assay (Functional)
            Assay-Dependent
            -
            Product Specifications
            Species Reactivity
            Mouse
            Published species
            Human, Mouse
            Host/Isotype
            Rat / IgG2a, kappa
            Class
            Monoclonal
            Type
            Antibody
            Clone
            AFL4
            Conjugate
            Unconjugated
            • Biotin 
            • Functional Grade
            • Request custom conjugation
            Form
            Liquid
            Concentration
            0.5 mg/mL
            Purification
            Affinity chromatography
            Storage buffer
            PBS, pH 7.2
            Contains
            0.09% sodium azide
            Storage conditions
            4°C
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            RRID
            AB_467796
            Product Specific Information
            Description: The AFL4 monoclonal antibody reacts with the mouse VEGF receptor-3, also known as Flt-4. This 195 kDa molecule was identified as an endothelial-specific member of the receptor tyrosine kinase (RTK) family. During early embryogenesis all endothelial cells express VEGFR-3, while in the adult tissues, VEGFR-3 expression disappears from the vascular endothelial cells and is observed only on the lymphatic endothelium. However, VEGFR-3 expression is induced in the adult tissue upon tumor implementation suggesting an important role for this receptor in the tumor angiogenesis. VEGF-C and VEGF-D bind to and activate VEGFR-3. AFL4 is an antagonist mAb.
            Applications Reported: The AFL4 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified AFL4, Product # 16-5988, in functional assays).
            Applications Tested: The AFL4 antibody has been tested by flow cytometric analysis of in vitro differentiated mouse endothelial cells. In brief, mouse ES cells were incubated on collagen IV matrix for 4 days and subsequently stimulated with VEGF under serum free conditions to induce further differentiation. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
            Purity: Greater than 90%, as determined by SDS-PAGE.
            Aggregation: Less than 10%, as determined by HPLC.
            Filtration: 0.2 µm post-manufacturing filtered.
            Target Information
            Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.
            Related Products for
            Immunohistochemistry
            • Secondary Antibodies
            • Additional Formats
            • Isotype Controls
            • Mounting Media IHC
            • Counter Stains IHC
            • Blockers IHC
            • Substrates IHC
            • Labeling Kits IHC
            Catalog #
            A21208
            Invitrogen
            Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
            Price (USD)
            341.00
            Unit Size :
            1 mg
            Catalog #
            A21434
            Invitrogen
            Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555
            Price (USD)
            290.00
            Unit Size :
            1 mg
            Catalog #
            A48272
            Invitrogen
            Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 647
            Price (USD)
            408.00
            Unit Size :
            1 mg
            Catalog #
            A21209
            Invitrogen
            Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594
            Unit Size :
            1 mg
            Catalog #
            31470
            Invitrogen
            Goat anti-Rat IgG (H+L) Secondary Antibody, HRP
            Unit Size :
            2 mL
            Catalog #
            A78945
            Invitrogen
            Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555
            Unit Size :
            1 mg
            Catalog #
            A11006
            Invitrogen
            Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
            Unit Size :
            1 mg
            Catalog #
            A11077
            Invitrogen
            Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 568
            Unit Size :
            1 mg
            Catalog #
            A21247
            Invitrogen
            Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647
            Unit Size :
            1 mg
            Catalog #
            A21094
            Invitrogen
            Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 633
            Unit Size :
            1 mg
            Bioinformatics
            Protein Aliases: chylous ascites; FLT-4; Fms-like tyrosine kinase 4; receptor protein tyrosine kinase; soluble VEGFR 3; sVEGFR 3; Tyrosine-protein kinase receptor FLT4; Vascular endothelial growth factor receptor 3; vascular endothelial growth factor receptor-3; VEGF R3; VEGFR-3
            View more View less
            Gene Aliases: AI323512; Chy; Flt-4; Flt4; VEGFR-3; VEGFR3
            View more View less
            UniProt ID: (Mouse) P35917
            View more View less
            Entrez Gene ID: (Mouse) 14257
            View more View less
            Function(s)
            nucleotide binding protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity vascular endothelial growth factor-activated receptor activity protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups growth factor binding protein phosphatase binding transmembrane signal receptor
            Process(es)
            angiogenesis positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation vasculature development lymph vessel development lymphangiogenesis sprouting angiogenesis protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cell proliferation positive regulation of vascular endothelial growth factor production positive regulation of endothelial cell migration phosphorylation peptidyl-tyrosine phosphorylation positive regulation of cell growth cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor signaling pathway negative regulation of apoptotic process positive regulation of MAPK cascade positive regulation of JNK cascade protein autophosphorylation vascular endothelial growth factor receptor signaling pathway blood vessel morphogenesis regulation of blood vessel remodeling positive regulation of ERK1 and ERK2 cascade positive regulation of protein kinase C signaling
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-7c4bf66445-tsvqc:80/100.66.79.38:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0