Transform your discoveries into market-ready therapies
Biotech solutions
    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • Histone H2A.X Antibodies
            Invitrogen
            Phospho-Histone H2A.X (Ser139) Monoclonal Antibody (CR55T33), eBioscience™
            Advanced Verification
            5 Published Figures
            7 References
            View all (76) Histone H2A.X antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 14-9865-80
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            Price (USD)
            120.65
            Online Exclusive
            161.00
            Save 40.35 (25%)
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers
            Cite Phospho-Histone H2A.X (Ser139) Monoclonal Antibody (CR55T33), eBioscience™
            • Antibody Testing Data (5)
            • Published Figures (5)
            • Advanced Verification (1)
            Application
            Phospho-Histone H2A.X (Ser139) Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.
            FIGURE: 1 / 11
            Phospho-Histone H2A.X (Ser139) Antibody (14-9865-80) in ICC/IF
            Immunofluorescence analysis of phospho-H2A.X was performed using 70% confluent log phase HeLa cells treated with hydroxyurea (3 mM for 6 hr). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Phospho-Histone H2A.X (Ser139) Monoclonal Antibody (CR55T33), eB... View More
            View Product
            Phospho-Histone H2A.X (Ser139) Antibody in Immunocytochemistry (ICC/IF)
            Phospho-Histone H2A.X (Ser139) Antibody in Immunocytochemistry (ICC/IF)
            Phospho-Histone H2A.X (Ser139) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Phospho-Histone H2A.X (Ser139) Antibody in Western Blot (WB)
            Phospho-Histone H2A.X (Ser139) Antibody in Flow Cytometry (Flow)
            Phospho-Histone H2A.X (Ser139) Antibody in Immunohistochemistry (IHC)
            Phospho-Histone H2A.X (Ser139) Antibody in Western Blot (WB)
            Phospho-Histone H2A.X (Ser139) Antibody in Western Blot (WB)
            Phospho-Histone H2A.X (Ser139) Antibody in Flow Cytometry (Flow)
            Phospho-Histone H2A.X (Ser139) Antibody in Flow Cytometry (Flow)
            Phospho-Histone H2A.X (Ser139) Antibody
            Product Details
            14-9865-80
            Applications
            Tested Dilution
            Publications
            Western Blot (WB)
            1:500
            View 2 publications 2 publications
            Immunohistochemistry (IHC)
            -
            View 1 publication 1 publication
            Immunohistochemistry (Paraffin) (IHC (P))
            5 µg/mL
            View 1 publication 1 publication
            Immunocytochemistry (ICC/IF)
            5 µg/mL
            View 1 publication 1 publication
            Flow Cytometry (Flow)
            0.03 µg/test
            View 2 publications 2 publications
            Product Specifications
            Species Reactivity
            Human, Mouse
            Published species
            Human
            Host/Isotype
            Mouse / IgG1, kappa
            Class
            Monoclonal
            Type
            Antibody
            Clone
            CR55T33
            Conjugate
            Unconjugated
            • Alexa Fluor 488 
            • eFluor 570
            • eFluor 660
            • PE 
            • PerCP-eFluor 710
            • Request custom conjugation
            Form
            Liquid
            Concentration
            0.5 mg/mL
            Purification
            Affinity chromatography
            Storage buffer
            PBS, pH 7.2
            Contains
            0.09% sodium azide
            Storage conditions
            4°C
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            RRID
            AB_2573047
            Product Specific Information
            Description: The CR55T33 monoclonal antibody recognizes phosphorylated serine 139 of human and mouse H2AX. H2AX is a member of the H2A histone family that complex with DNA and other histones to form the repeating nucleosome units characteristic of eukaryotic chromatin. Nucleosomes consist of approximately 147 base pairs of DNA wrapped around an octamer of histones composed of two each of the four histone proteins: H2A, H2B, H3 and H4. After induction of DNA damage such as double-strand breaks by irradiation, genotoxic stresses, replication errors or gene recombination, PI3K-like kinases (e.g., ataxia telangiectasia mutated (ATM), ataxia telangiectasia Rad-3-related (ATR), and DNA-dependent protein kinase (DNA-PK) are activated to phosphorylate serine 139 in H2AX. This early phosphorylation event plays a critical role in recruiting proteins involved in DNA repair.
            The monoclonal antibody CR55T33 recognizes a single band of approximately 15 kDa on reduced cell lysates from Jurkat cells stimulated with etoposide.
            Applications Reported: This CR55T33 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry (fluorochrome-conjugated CR55T33 is recommended for use in intracellular flow cytometry).
            Applications Tested: The CR55T33 has been tested by immunocytochemistry of methanol-fixed cells and by immunohistochemistry of human tissue using either low or high pH antigen retrieval and can be used at 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This CR55T33 antibody has also been tested by intracellular staining and flow cytometric analysis of stimulated Jurkat cells using the Foxp3/Transcription Factor Buffer Set (Product # 00-5523-00) and protocol. This can be used at 0.03 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
            Protocols: We recommend Protocol B: One-step protocol: intracellular (nuclear) proteins. Alternatively, Protocol C: Two-step protocol: Fixation/Methanol can also be used. Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins cannot be used. All Protocols can be found in the "Staining intracellular Antigens for Flow Cytometry Protocol" located in the BestProtocols® Section under the Resources tab online.
            eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome.
            Purity: Greater than 90%, as determined by SDS-PAGE.
            Aggregation: Less than 10%, as determined by HPLC.
            Filtration: 0.2 µm post-manufacturing filtered.
            Target Information
            Histone H2A.X (H2AX) is a member of the histone H2A family which is one of the four core histones making up the nucleosome core particle. In eukaryotes, DNA double strand breaks (DSBs) have been shown to trigger the phosphorylation of serine 139 at the carboxy terminus of histone H2AX resulting in gamma-H2AX. The phosphorylation of H2AX can be detected by Western blotting or immunofluorescence, revealing the frequency of DSBs. The phosphatidylinositol 3-kinases have been implicated in H2AX phosphorylation, but it is unclear if ATM is the primary H2AX kinase or if other members of the family such as DNA-PK and ATR contribute in a similar manner. Structurally, H2A.x contains 143 amino acid residues. Histone H2A.X is considered a basal histone, being synthesized in G1 as well as in S-phase, and its mRNA contains polyA addition motifs and a polyA tail along with the conserved stem-loop and U7 binding sequences involved in the processing and stability of replication type histone mRNAs. There are two forms of Histone H2A.X mRNA, one about 1600 bases long and contains polyA; the other about 575 bases long, lacking polyA. The short form behaves as a replication type histone mRNA, while the longer behaves as a basal type histone mRNA. Histone H2A.X maps to the 11q23.2-q23.3 region of the human chromosome. Histone H2A.x contributes to histone-formation and therefore the structure of DNA. Histone H2A variant H2A.x specifically regulates the interaction of MDC1 (mediator of DNA damage checkpoint protein 1), a DNA repair protein to the sites of DNA damage.
            Related Products for
            Western Blot
            • Secondary Antibodies
            • Additional Formats
            • Isotype Controls
            • Loading Controls WB
            • Electrophoresis WB
            • Transfer Stacks WB
            • Substrates WB
            • Labeling Kits WB
            Catalog #
            31160
            Invitrogen
            Goat anti-Mouse IgG (H+L) Secondary Antibody
            Unit Size :
            2 mg
            Catalog #
            31800
            Invitrogen
            Goat anti-Mouse IgG (H+L) Secondary Antibody, Biotin
            Unit Size :
            2 mL
            Catalog #
            A31571
            Invitrogen
            Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647
            Unit Size :
            1 mg
            Catalog #
            A21200
            Invitrogen
            Chicken anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
            Unit Size :
            1 mg
            Catalog #
            A21203
            Invitrogen
            Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594
            Unit Size :
            1 mg
            Catalog #
            A21202
            Invitrogen
            Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488
            Unit Size :
            1 mg
            Catalog #
            616520
            Invitrogen
            Rabbit anti-Mouse IgG (H+L) Secondary Antibody, HRP
            Unit Size :
            1 mL
            Catalog #
            A10036
            Invitrogen
            Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 546
            Unit Size :
            1 mg
            Catalog #
            A32728
            Invitrogen
            Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 647
            Unit Size :
            1 mg
            Catalog #
            A21037
            Invitrogen
            Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 750
            Unit Size :
            1 mg
            Bioinformatics
            Protein Aliases: gamma H2AX; H2A histone family, member X; H2a/x; H2AX histone; histone 5 protein 2ax; Histone H2A.X; Histone H2a/x; Histone H2AX
            View more View less
            Gene Aliases: AW228881; gammaH2ax; H2A.X; H2A/X; H2AFX; H2AX; Hist5-2ax
            View more View less
            UniProt ID: (Human) P16104, (Mouse) P27661
            View more View less
            Entrez Gene ID: (Human) 3014, (Mouse) 15270
            View more View less
            Function(s)
            DNA binding damaged DNA binding protein binding enzyme binding histone binding protein heterodimerization activity histone
            Process(es)
            DNA damage checkpoint double-strand break repair via homologous recombination double-strand break repair double-strand break repair via nonhomologous end joining nucleosome assembly chromatin silencing cellular response to DNA damage stimulus spermatogenesis response to ionizing radiation viral process cerebral cortex development positive regulation of DNA repair meiotic cell cycle cellular response to gamma radiation cellular senescence DNA repair DNA recombination cell cycle
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-7c4bf66445-q5dkm:80/100.66.74.143:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0