Product References
The Bisdioxopiperazine ICRF-193 Attenuates LPS-induced IL-1β Secretion by Macrophages.
Inflammation
Brindle A,Bainbridge C,Kumar MR,Todryk S,Padget K
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Myeloid‑derived suppressor cell accumulation induces Treg expansion and modulates lung malignancy progression.
Biomedical reports
Wan Y,Mu X,Zhao J,Li L,Xu W,Zhang M
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
ELMO1 Deficiency Reduces Neutrophil Chemotaxis in Murine Peritonitis.
International journal of molecular sciences
Yu S,Geng X,Liu H,Zhang Y,Cao X,Li B,Yan J
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sun Apr 30 00:00:00 UTC 2023
Regulation of synaptic connectivity in schizophrenia spectrum by mutual neuron-microglia interaction.
Communications biology
Breitmeyer R,Vogel S,Heider J,Hartmann SM,Wüst R,Keller AL,Binner A,Fitzgerald JC,Fallgatter AJ,Volkmer H
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sat Apr 29 00:00:00 UTC 2023
Local Myeloid-Derived Suppressor Cells Impair Progression of Experimental Autoimmune Uveitis by Alleviating Oxidative Stress and Inflammation.
Investigative ophthalmology & visual science
Lee JY,Kim S,Sohn HJ,Kim CH,Kim TG,Lee HS
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Oct 03 00:00:00 UTC 2023
LAL deficiency induced myeloid-derived suppressor cells as targets and biomarkers for lung cancer.
Journal for immunotherapy of cancer
Zhao T,Liu S,Hanna NH,Jalal S,Ding X,Wan J,Yan C,Du H
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2023
Methylation of SPRED1: A New Target in Acute Myeloid Leukemia.
Frontiers in oncology
Su N,Wang Y,Lu X,Xu W,Wang H,Mo W,Pang H,Tang R,Li S,Yan X,Li Y,Zhang R
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sat Apr 02 00:00:00 UTC 2022
Prognostic Lnc-S100B-2 Affects Cell Apoptosis and Microenvironment of Colorectal Cancer through MLLT10 Signaling.
Journal of oncology
Yi J,Peng F,Zhao J,Gong X
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Feb 08 00:00:00 UTC 2022
Pharmacological inhibition of LSD1 triggers myeloid differentiation by targeting GSE1 oncogenic functions in AML.
Oncogene
Nicosia L,Boffo FL,Ceccacci E,Conforti F,Pallavicini I,Bedin F,Ravasio R,Massignani E,Somervaille TCP,Minucci S,Bonaldi T
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Feb 01 00:00:00 UTC 2022
miR-204-containing exosomes ameliorate GVHD-associated dry eye disease.
Science advances
Zhou T,He C,Lai P,Yang Z,Liu Y,Xu H,Lin X,Ni B,Ju R,Yi W,Liang L,Pei D,Egwuagu CE,Liu X
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Fri Jan 14 00:00:00 UTC 2022
The Transcription of ZIP9 Is Associated With the Macrophage Polarization and the Pathogenesis of Hepatocellular Carcinoma.
Frontiers in immunology
Gou Y,Yang D,Tian T,Zhu X,Zhang R,Ren J,Tu D,Luo Y,Miao Y,Zhao H,Wang Y,Wei B
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Fri Apr 08 00:00:00 UTC 2022
Incomplete penetrance of NOD2 C483W mutation underlining Blau syndrome.
Pediatric rheumatology online journal
Chang SY,Kambe N,Fan WL,Huang JL,Lee WI,Wu CY
15-0118-42 was used in Flow cytometry/Cell sorting to report a novel C483W NOD2 mutation underlining BS with incomplete penetrance.
Mon Oct 03 00:00:00 UTC 2022
Lysosomal acid lipase, CSF1R, and PD-L1 determine functions of CD11c+ myeloid-derived suppressor cells.
JCI insight
Zhao T,Liu S,Ding X,Johnson EM,Hanna NH,Singh K,Sen CK,Wan J,Du H,Yan C
15-0118-42 was used in Flow cytometry/Cell sorting to investigate if CD11c+ cells switched their functions to immune suppression and tumor growth stimulation through CSF1R/PD-L1 upregulation and metabolic reprogramming.
Thu Sep 08 00:00:00 UTC 2022
Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
The Journal of experimental medicine
Louka E,Povinelli B,Rodriguez-Meira A,Buck G,Wen WX,Wang G,Sousos N,Ashley N,Hamblin A,Booth CAG,Roy A,Elliott N,Iskander D,de la Fuente J,Fordham N,O'Byrne S,Inglott S,Norfo R,Salio M,Thongjuea S,Rao A,Roberts I,Mead AJ
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Different Induction of PD-L1 (CD274) and PD-1 (CD279) Expression in THP-1-Differentiated Types 1 and 2 Macrophages.
Journal of inflammation research
Lai CY,Tseng PC,Chen CL,Satria RD,Wang YT,Lin CF
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Wed Apr 27 00:00:00 UTC 2022
G-MDSCs promote aging-related cardiac fibrosis by activating myofibroblasts and preventing senescence.
Cell death & disease
Sun SN,Ni SH,Li Y,Liu X,Deng JP,Chen ZX,Li H,Feng WJ,Huang YS,Li DN,Xian SX,Yang ZQ,Wang LJ,Lu L
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
Fat-1 Transgenic Mice With Augmented n3-Polyunsaturated Fatty Acids Are Protected From Liver Injury Caused by Acute-On-Chronic Ethanol Administration.
Frontiers in pharmacology
Warner J,Hardesty J,Song Y,Sun R,Deng Z,Xu R,Yin X,Zhang X,McClain C,Warner D,Kirpich I
15-0118-42 was used in Flow Cytometry to interrogate the effects of endogenous n3-PUFA enrichment in a mouse model which recapitulates features of early human alcohol-associated hepatitis.
Thu Sep 29 00:00:00 UTC 2022
Associations between Hypertriglyceridemia and Circulating Neutrophil Subpopulation in Patients with Dyslipidemia.
International journal of inflammation
Genkel V,Dolgushin I,Baturina I,Savochkina A,Kuznetsova A,Pykhova L,Shaposhnik I
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sun Apr 24 00:00:00 UTC 2022
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu May 07 00:00:00 UTC 2020
The long non-coding RNA Cancer Susceptibility 15 (CASC15) is induced by isocitrate dehydrogenase (IDH) mutations and maintains an immature phenotype in adult acute myeloid leukemia.
Haematologica
Grasedieck S,Ruess C,Krowiorz K,Lux S,Pochert N,Schwarzer A,Klusmann JH,Jongen-Lavrencic M,Herold T,Bullinger L,Pollack JR,Rouhi A,Kuchenbauer F
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Galectin-9 promotes a suppressive microenvironment in human cancer by enhancing STING degradation.
Oncogenesis
Zhang CX,Huang DJ,Baloche V,Zhang L,Xu JX,Li BW,Zhao XR,He J,Mai HQ,Chen QY,Zhang XS,Busson P,Cui J,Li J
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Jul 06 00:00:00 UTC 2020
Connecting METTL3 and intratumoural CD33+ MDSCs in predicting clinical outcome in cervical cancer.
Journal of translational medicine
Ni HH,Zhang L,Huang H,Dai SQ,Li J
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Oct 15 00:00:00 UTC 2020
Ubiquitin-specific peptidase 3 induces TPA-mediated leukemia cell differentiation via regulating H2AK119ub.
Animal cells and systems
Chae YC,Jung H,Kim JY,Lee DH,Seo SB
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Sep 29 00:00:00 UTC 2020
Microbiome Influences Prenatal and Adult Microglia in a Sex-Specific Manner.
Cell
Thion MS,Low D,Silvin A,Chen J,Grisel P,Schulte-Schrepping J,Blecher R,Ulas T,Squarzoni P,Hoeffel G,Coulpier F,Siopi E,David FS,Scholz C,Shihui F,Lum J,Amoyo AA,Larbi A,Poidinger M,Buttgereit A,Lledo PM,Greter M,Chan JKY,Amit I,Beyer M,Schultze JL,Schlitzer A,Pettersson S,Ginhoux F,Garel S
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Jan 25 00:00:00 UTC 2018
AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
Molecules and cells
Park JW,Cho H,Oh H,Kim JY,Seo SB
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu May 31 00:00:00 UTC 2018
n-butanol extract from Folium isatidis inhibits the lipopolysaccharide-induced downregulation of CXCR1 and CXCR2 on human neutrophils.
Molecular medicine reports
Wu B,Wang L,Jiang L,Dong L,Xu F,Lu Y,Jin J,Wang Z,Liang G,Shan X
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
Deacetylase activity-independent transcriptional activation by HDAC2 during TPA-induced HL-60 cell differentiation.
PloS one
Jung H,Kim JY,Kim KB,Chae YC,Hahn Y,Kim JW,Seo SB
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Feb 07 00:00:00 UTC 2019
Microglia innately develop within cerebral organoids.
Nature communications
Ormel PR,Vieira de Sá R,van Bodegraven EJ,Karst H,Harschnitz O,Sneeboer MAM,Johansen LE,van Dijk RE,Scheefhals N,Berdenis van Berlekom A,Ribes Martínez E,Kling S,MacGillavry HD,van den Berg LH,Kahn RS,Hol EM,de Witte LD,Pasterkamp RJ
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Oct 09 00:00:00 UTC 2018
The prognostic significance of CD11b+CX3CR1+ monocytes in patients with newly diagnosed diffuse large B-cell lymphoma.
Oncotarget
Yhim HY,Kim JA,Ko SH,Park Y,Yim E,Kim HS,Kwak JY
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Fri Nov 03 00:00:00 UTC 2017
CD19 CAR immune pressure induces B-precursor acute lymphoblastic leukaemia lineage switch exposing inherent leukaemic plasticity.
Nature communications
Jacoby E,Nguyen SM,Fountaine TJ,Welp K,Gryder B,Qin H,Yang Y,Chien CD,Seif AE,Lei H,Song YK,Khan J,Lee DW,Mackall CL,Gardner RA,Jensen MC,Shern JF,Fry TJ
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Wed Jul 27 00:00:00 UTC 2016
Metabolic regulation of hepatitis B immunopathology by myeloid-derived suppressor cells.
Nature medicine
Pallett LJ,Gill US,Quaglia A,Sinclair LV,Jover-Cobos M,Schurich A,Singh KP,Thomas N,Das A,Chen A,Fusai G,Bertoletti A,Cantrell DA,Kennedy PT,Davies NA,Haniffa M,Maini MK
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
STK4 regulates TLR pathways and protects against chronic inflammation-related hepatocellular carcinoma.
The Journal of clinical investigation
Li W,Xiao J,Zhou X,Xu M,Hu C,Xu X,Lu Y,Liu C,Xue S,Nie L,Zhang H,Li Z,Zhang Y,Ji F,Hui L,Tao W,Wei B,Wang H
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Mon Nov 02 00:00:00 UTC 2015
Induction of autophagy is a key component of all-trans-retinoic acid-induced differentiation in leukemia cells and a potential target for pharmacologic modulation.
Experimental hematology
Orfali N,O'Donovan TR,Nyhan MJ,Britschgi A,Tschan MP,Cahill MR,Mongan NP,Gudas LJ,McKenna SL
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Tue Sep 15 00:00:00 UTC 2015
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) preserves gut barrier function by blocking neutrophil-endothelial cell adhesion after hemorrhagic shock and resuscitation in mice.
Surgery
Zhang HY,James I,Chen CL,Besner GE
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2012
Normal human monocytes exposed to glioma cells acquire myeloid-derived suppressor cell-like properties.
Neuro-oncology
Rodrigues JC,Gonzalez GC,Zhang L,Ibrahim G,Kelly JJ,Gustafson MP,Lin Y,Dietz AB,Forsyth PA,Yong VW,Parney IF
Published figure using CD11b monoclonal antibody (Product # 15-0118-42) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2010