Product References
Activated CD4+ T cells and CD14hiCD16+ monocytes correlate with antibody response following influenza virus infection in humans.
Cell reports. Medicine
Wong SS,Oshansky CM,Guo XJ,Ralston J,Wood T,Reynolds GE,Seeds R,Jelley L,Waite B,Jeevan T,Zanin M,Widdowson MA,Huang QS,Thomas PG,Webby RJ
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Apr 20 00:00:00 UTC 2021
The role of irreversible electroporation in promoting M1 macrophage polarization via regulating the HMGB1-RAGE-MAPK axis in pancreatic cancer.
Oncoimmunology
He C,Sun S,Zhang Y,Xie F,Li S
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Thu Mar 11 00:00:00 UTC 2021
Transplantation of human amniotic epithelial cells promotes morphological and functional regeneration in a rat uterine scar model.
Stem cell research & therapy
Fan Y,Sun J,Zhang Q,Lai D
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Wed Mar 24 00:00:00 UTC 2021
Monitoring of Peripheral Blood Leukocytes and Plasma Samples: A Pilot Study to Examine Treatment Response to Leflunomide in Rheumatoid Arthritis.
Pharmaceuticals (Basel, Switzerland)
Rodrigues JFS,da Silva LCM,Cardoso-Sousa L,Caixeta DC,Lückemeyer DD,Henrique AS,Pontes JP,da Silva LMG,Macedo JSS,Carvalho Júnior PS,Silva E Silva C,Martins MMRS,Monteiro-Neto V,Grisotto MAG,Fernandes AMR,Ferreira J,Calixto JB,Sabino-Silva R,Fernandes ES
15-9956-42 was used in Flow Cytometry to investigate the potential use of flow cytometry and attenuated total reflection-Fourier transform infrared (ATR-FTIR) spectroscopy as measures to identify responders and non-responders to leflunomide, a disease-modifying drug used in the treatment of patients with rheumatoid arthritis.
Fri Jan 29 00:00:00 UTC 2021
Lung transplantation for patients with severe COVID-19.
Science translational medicine
Bharat A,Querrey M,Markov NS,Kim S,Kurihara C,Garza-Castillon R,Manerikar A,Shilatifard A,Tomic R,Politanska Y,Abdala-Valencia H,Yeldandi AV,Lomasney JW,Misharin AV,Budinger GRS
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Wed Dec 16 00:00:00 UTC 2020
Gene expression network analyses during infection with virulent and avirulent Trypanosoma cruzi strains unveil a role for fibroblasts in neutrophil recruitment and activation.
PLoS pathogens
Oliveira AER,Pereira MCA,Belew AT,Ferreira LRP,Pereira LMN,Neves EGA,Nunes MDCP,Burleigh BA,Dutra WO,El-Sayed NM,Gazzinelli RT,Teixeira SMR
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Characterization of HLA-G Regulation and HLA Expression in Breast Cancer and Malignant Melanoma Cell Lines upon IFN-γ Stimulation and Inhibition of DNA Methylation.
International journal of molecular sciences
Jørgensen N,Sayed A,Jeppesen HB,Persson G,Weisdorf I,Funck T,Hviid TVF
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Wed Jun 17 00:00:00 UTC 2020
Connecting METTL3 and intratumoural CD33+ MDSCs in predicting clinical outcome in cervical cancer.
Journal of translational medicine
Ni HH,Zhang L,Huang H,Dai SQ,Li J
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Thu Oct 15 00:00:00 UTC 2020
New GMP manufacturing processes to obtain thermostable HIV-1 gp41 virosomes under solid forms for various mucosal vaccination routes.
NPJ vaccines
Amacker M,Smardon C,Mason L,Sorrell J,Jeffery K,Adler M,Bhoelan F,Belova O,Spengler M,Punnamoottil B,Schwaller M,Bonduelle O,Combadière B,Stegmann T,Naylor A,Johnson R,Wong D,Fleury S
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Fri Dec 17 00:00:00 UTC 2021
Targeting the mTOR pathway uncouples the efficacy and toxicity of PD-1 blockade in renal transplantation.
Nature communications
Esfahani K,Al-Aubodah TA,Thebault P,Lapointe R,Hudson M,Johnson NA,Baran D,Bhulaiga N,Takano T,Cailhier JF,Piccirillo CA,Miller WH
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Thu Oct 17 00:00:00 UTC 2019
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
Large-Scale Human Dendritic Cell Differentiation Revealing Notch-Dependent Lineage Bifurcation and Heterogeneity.
Cell reports
Balan S,Arnold-Schrauf C,Abbas A,Couespel N,Savoret J,Imperatore F,Villani AC,Vu Manh TP,Bhardwaj N,Dalod M
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Aug 14 00:00:00 UTC 2018
Role of Nuclear Factor (Erythroid-Derived 2)-Like 2 Signaling for Effects of Fumaric Acid Esters on Dendritic Cells.
Frontiers in immunology
Hammer A,Waschbisch A,Knippertz I,Zinser E,Berg J,Jörg S,Kuhbandner K,David C,Pi J,Bayas A,Lee DH,Haghikia A,Gold R,Steinkasserer A,Linker RA
15-9956 was used in Flow cytometry/Cell sorting to reveal, using treatment with fumaric acid esters as a model, the effects of Nrf2 activation and signalling on dendritic cells.
Wed Nov 20 00:00:00 UTC 2019
HDAC inhibition potentiates immunotherapy in triple negative breast cancer.
Oncotarget
Terranova-Barberio M,Thomas S,Ali N,Pawlowska N,Park J,Krings G,Rosenblum MD,Budillon A,Munster PN
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Dec 26 00:00:00 UTC 2017
Gut-homing Δ42PD1+Vδ2 T cells promote innate mucosal damage via TLR4 during acute HIV type 1 infection.
Nature microbiology
Cheung AKL,Kwok HY,Huang Y,Chen M,Mo Y,Wu X,Lam KS,Kong HK,Lau TCK,Zhou J,Li J,Cheng L,Kiat Lee B,Peng Q,Lu X,An M,Wang H,Shang H,Zhou B,Wu H,Xu A,Yuen KY,Chen Z
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2017
Surrogate in vitro activation of innate immunity synergizes with interleukin-7 to unleash rapid antigen-driven outgrowth of CD4+ and CD8+ human peripheral blood T-cells naturally recognizing MUC1, HER2/neu and other tumor-associated antigens.
Oncotarget
Pathangey LB,McCurry DB,Gendler SJ,Dominguez AL,Gorman JE,Pathangey G,Mihalik LA,Dang Y,Disis ML,Cohen PA
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Feb 14 00:00:00 UTC 2017
FOXP1 suppresses immune response signatures and MHC class II expression in activated B-cell-like diffuse large B-cell lymphomas.
Leukemia
Brown PJ,Wong KK,Felce SL,Lyne L,Spearman H,Soilleux EJ,Pedersen LM,Møller MB,Green TM,Gascoyne DM,Banham AH
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Lack of T Cell Response to iPSC-Derived Retinal Pigment Epithelial Cells from HLA Homozygous Donors.
Stem cell reports
Sugita S,Iwasaki Y,Makabe K,Kimura T,Futagami T,Suegami S,Takahashi M
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Oct 11 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using HLA-DR monoclonal antibody (Product # 15-9956-42) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
Lipopolysaccharide regulation of toll-like receptor-4 and matrix metalloprotease-9 in human primary corneal fibroblasts.
Investigative ophthalmology & visual science
Wong Y,Sethu C,Louafi F,Hossain P
15-9956 was used in Flow cytometry/Cell sorting to show that TLR4 activation with LPS increases TLR4, MMP-9, and cytokine expression in CFs cultured from patients with microbial keratitis.
Mon Apr 25 00:00:00 UTC 2011