Product References
USP7 inhibits the progression of nasopharyngeal carcinoma via promoting SPLUNC1-mediated M1 macrophage polarization through TRIM24.
Cell death & disease
Liu H,Tang L,Gong S,Xiao T,Yang H,Gu W,Wang H,Chen P
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Thu Dec 21 00:00:00 UTC 2023
Lipoxins modulate neutrophil oxidative burst, integrin expression and lymphatic transmigration differentially in human health and atherosclerosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Kraft JD,Blomgran R,Bergström I,Soták M,Clark M,Rani A,Rajan MR,Dalli J,Nyström S,Quiding-Järbrink M,Bromberg J,Skoog P,Börgeson E
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2022
Upregulation of CD3ζ and L-selectin in antigen-specific cytotoxic T lymphocytes by eliminating myeloid-derived suppressor cells with doxorubicin to improve killing efficacy of neuroblastoma cells in vitro.
Journal of clinical laboratory analysis
Xu W,Li S,Li M,Zhou H,Yang X
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Sat Jan 01 00:00:00 UTC 2022
Neutrophil activation and circulating neutrophil extracellular traps are increased in venous thromboembolism patients for at least one year after the clinical event.
Journal of thrombosis and thrombolysis
Zapponi KCS,Orsi FA,Cunha JLR,de Brito IR,Romano AVC,Bittar LF,De Paula EV,Penteado CF,Montalvão S,Annichino-Bizzacchi JM
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Sat Jan 01 00:00:00 UTC 2022
Bletinib ameliorates neutrophilic inflammation and lung injury by inhibiting Src family kinase phosphorylation and activity.
British journal of pharmacology
Kao TI,Chen PJ,Wang YH,Tseng HH,Chang SH,Wu TS,Yang SH,Lee YT,Hwang TL
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Fri Oct 01 00:00:00 UTC 2021
RAS-Responsive Element-Binding Protein 1 Blocks the Granulocytic Differentiation of Myeloid Leukemia Cells.
Oncology research
Yao J,Zhong L,Zhong P,Liu D,Yuan Z,Liu J,Yao S,Zhao Y,Chen M,Li L,Liu L,Liu B
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Fri Jul 12 00:00:00 UTC 2019
Deficiency of FAM3D (Family With Sequence Similarity 3, Member D), A Novel Chemokine, Attenuates Neutrophil Recruitment and Ameliorates Abdominal Aortic Aneurysm Development.
Arteriosclerosis, thrombosis, and vascular biology
He L,Fu Y,Deng J,Shen Y,Wang Y,Yu F,Xie N,Chen Z,Hong T,Peng X,Li Q,Zhou J,Han J,Wang Y,Xi J,Kong W
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
Impact of combined ischemic preconditioning and remote ischemic perconditioning on ischemia-reperfusion injury after liver transplantation.
Scientific reports
Li DY,Liu WT,Wang GY,Shi XJ
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Wed Dec 19 00:00:00 UTC 2018
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Tue Jan 19 00:00:00 UTC 2016
NLS‑RARα modulates acute promyelocytic leukemia NB4 cell proliferation and differentiation via the PI3K/AKT pathway.
Molecular medicine reports
Song H,Li L,Zhong L,Yang R,Jiang K,Yang X,Liu B
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2016
CD16 is indispensable for antibody-dependent cellular cytotoxicity by human monocytes.
Scientific reports
Yeap WH,Wong KL,Shimasaki N,Teo EC,Quek JK,Yong HX,Diong CP,Bertoletti A,Linn YC,Wong SC
16-0113 was used in Flow cytometry/Cell sorting to show CD16+ monocytes are important effectors of ADCC, potentially significant for cellular therapies for cancer and infectious diseases.
Tue Sep 27 00:00:00 UTC 2016
Serum biomarkers VEGF-C and IL-6 are associated with severe human Peripheral Artery Stenosis.
Journal of inflammation (London, England)
Chen J,Han L,Xu X,Tang H,Wang H,Wei B
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Tue Aug 18 00:00:00 UTC 2015
MIR125B1 represses the degradation of the PML-RARA oncoprotein by an autophagy-lysosomal pathway in acute promyelocytic leukemia.
Autophagy
Zeng CW,Chen ZH,Zhang XJ,Han BW,Lin KY,Li XJ,Wei PP,Zhang H,Li Y,Chen YQ
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Wed Oct 01 00:00:00 UTC 2014
Human scavenger protein AIM increases foam cell formation and CD36-mediated oxLDL uptake.
Journal of leukocyte biology
Amézaga N,Sanjurjo L,Julve J,Aran G,Pérez-Cabezas B,Bastos-Amador P,Armengol C,Vilella R,Escolà-Gil JC,Blanco-Vaca F,Borràs FE,Valledor AF,Sarrias MR
16-0113 was used in Flow cytometry/Cell sorting to study how hAIM is involved in macrophage survival, adhesion, and foam cell formation, and can serve to facilitate CD36-mediated uptake of oxLDL.
Sat Mar 01 00:00:00 UTC 2014
5-Lipoxygenase plays a pivotal role in endothelial adhesion of monocytes via an increased expression of Mac-1.
Cardiovascular research
Lee SJ,Choi EK,Seo KW,Bae JU,Kim YH,Park SY,Oh SO,Kim CD
16-0113 was used in Inhibition experiments to investigate the mechanism responsible for 5-LO expression in monocytes and its role in monocyte adhesion to the vascular endothelium.
Sun Sep 01 00:00:00 UTC 2013
5-Lipoxygenase plays a pivotal role in endothelial adhesion of monocytes via an increased expression of Mac-1.
Cardiovascular research
Lee SJ,Choi EK,Seo KW,Bae JU,Kim YH,Park SY,Oh SO,Kim CD
16-0113 was used in Inhibition experiments to investigate the mechanism responsible for 5-LO expression in monocytes and its role in monocyte adhesion to the vascular endothelium.
Sun Sep 01 00:00:00 UTC 2013
Complement receptor 3 influences toll-like receptor 7/8-dependent inflammation: implications for autoimmune diseases characterized by antibody reactivity to ribonucleoproteins.
The Journal of biological chemistry
Reed JH,Jain M,Lee K,Kandimalla ER,Faridi MH,Buyon JP,Gupta V,Clancy RM
16-0113 was used in Functional assays to investigate whether activation of CR3 negatively regulates TLR7/8-dependent effector function.
Fri Mar 29 00:00:00 UTC 2013
Gu-4 suppresses affinity and avidity modulation of CD11b and improves the outcome of mice with endotoxemia and sepsis.
PloS one
Yan T,Li Q,Zhou H,Zhao Y,Yu S,Xu G,Yin Z,Li Z,Zhao Z
Published figure using CD11b (activation epitope) monoclonal antibody (Product # 16-0113-82) in Flow Cytometry
Mon Sep 24 00:00:00 UTC 2012
Identification of endothelial cell junctional proteins and lymphocyte receptors involved in transendothelial migration of human effector memory CD4+ T cells.
Journal of immunology (Baltimore, Md. : 1950)
Manes TD,Pober JS
16-0113 was used in Blocking experiments to further analyze effector memory (EM) CD4(+) T cell migrational responses to either chemokine or TCR-activating signals.
Tue Feb 01 00:00:00 UTC 2011
A systemic lupus erythematosus-associated R77H substitution in the CD11b chain of the Mac-1 integrin compromises leukocyte adhesion and phagocytosis.
The Journal of biological chemistry
MacPherson M,Lek HS,Prescott A,Fagerholm SC
16-0113 was used in Flow cytometry/Cell sorting to investigate the effect of R77H-substituted Mac-1 on leukocytes.
Fri May 13 00:00:00 UTC 2011
Inflamed lymphatic endothelium suppresses dendritic cell maturation and function via Mac-1/ICAM-1-dependent mechanism.
Journal of immunology (Baltimore, Md. : 1950)
Podgrabinska S,Kamalu O,Mayer L,Shimaoka M,Snoeck H,Randolph GJ,Skobe M
16-0113 was used in Flow cytometry/Cell sorting to demonstrate that lymphatic endothelial cells modulate the maturation and function of dendritic cells.
Sat Aug 01 00:00:00 UTC 2009
High-mobility group box 1 activates integrin-dependent homing of endothelial progenitor cells.
Circulation research
Chavakis E,Hain A,Vinci M,Carmona G,Bianchi ME,Vajkoczy P,Zeiher AM,Chavakis T,Dimmeler S
16-0113 was used in Neutralization experiments to establish a link between HMGB1 and integrin functions of EPCs and demonstrate that HMGB1 stimulates EPC homing to ischemic tissues.
Fri Feb 02 00:00:00 UTC 2007
Porphyromonas gingivalis fimbriae proactively modulate beta2 integrin adhesive activity and promote binding to and internalization by macrophages.
Infection and immunity
Hajishengallis G,Wang M,Harokopakis E,Triantafilou M,Triantafilou K
16-0113 was used in Binding experiments to show that P. gingivalis fimbriae function as both an activator and a ligand of CD11b/CD18.
Sun Oct 01 00:00:00 UTC 2006