    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • CD16 Antibodies
            Invitrogen
            CD16 Monoclonal Antibody (B73.1), Functional Grade, eBioscience™
            2 Published Figures
            8 References
            View all (184) CD16 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 16-0167-85
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            Price (USD)
            481.10
            Special offer
            Online exclusive
            Ends: 01-Jul-2025
            566.00
            Save 84.90 (15%)
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers
            Cite CD16 Monoclonal Antibody (B73.1), Functional Grade, eBioscience™
            • Published Figures (2)
            Application
            CD16 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.
            FIGURE: 1 / 2
            CD16 Antibody (16-0167-85) in WB
            European journal of immunology 2009 - 5 PI3K and GSK-3beta function to inhibit TNF-alpha release. (A) Cultured human primary monocytes were pretreated with PI3K kinase inhibitor LY294002 or GSK-3beta kinase inhibitor LiCl 1 h prior to the addition of anti-CD16 antibody (clone 3G8) or vehicle. Monocytes were collected 0, 15 or 60 min after the addition of the CD16 antibody or vehicle, and 10 mug of total protein was loaded in each lane for Weste... View More
            View Product
            CD16 Antibody in Western Blot (WB)
            CD16 Antibody in Flow Cytometry (Flow)
            Product Details
            16-0167-85
            Applications
            Tested Dilution
            Publications
            Western Blot (WB)
            -
            View 1 publication 1 publication
            Flow Cytometry (Flow)
            1 µg/test
            View 4 publications 4 publications
            Neutralization (Neu)
            Assay-Dependent
            View 2 publications 2 publications
            Functional Assay (Functional)
            Assay-Dependent
            -
            Miscellaneous PubMed (Misc)
            -
            View 1 publication 1 publication
            Product Specifications
            Species Reactivity
            Human
            Published species
            Human
            Host/Isotype
            Mouse / IgG1, kappa
            Recommended Isotype Control
            Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Functional Grade, eBioscience™
            Class
            Monoclonal
            Type
            Antibody
            Clone
            B73.1
            Conjugate
            Functional Grade
            • PE 
            • Request custom conjugation
            Form
            Liquid
            Concentration
            1 mg/mL
            Purification
            Affinity chromatography
            Storage buffer
            PBS, pH 7.2
            Contains
            no preservative
            Storage conditions
            4°C
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            RRID
            AB_11040983
            Product Specific Information
            Description: This B73.1 monoclonal antibody reacts with human CD16, a member of the Fc receptor family also known as Fc gamma RIIIa and Fc gamma RIIIb. This 50-65 kDa receptor is expressed on natural killer cells, neutrophils, monocytes, and macrophages. Crosslinking of CD16 in natural killer cells and monocytes results in activation of the MAPK and PI3K signaling pathways, thus regulating the production of TNF alpha, IL-6, and IL-1 beta.
            This monoclonal antibody has been shown to block Fc receptor functions.
            Applications Reported: This B73.1 antibody has been reported for use in functional blocking assays and flow cytometric analysis.
            Applications Tested: This B73.1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
            Storage and handling: Use in a sterile environment.
            Filtration: 0.2 µm post-manufacturing filtered.
            Purity: Greater than 90%, as determined by SDS-PAGE.
            Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
            Aggregation: Less than 10%, as determined by HPLC.
            Target Information
            CD16 encodes a receptor that recognizes the Fc portion of immunoglobulin G and is involved in the clearance of immune complexes from the circulation, as well as other functions such as cellular mediated cytotoxicity and enhancement of virus infections. This gene, FCGR3A, shares a high degree of similarity with another nearby gene, FCGR3B, located on chromosome 1. The receptor encoded by this gene is expressed on natural killer (NK) cells as an integral membrane glycoprotein anchored through a transmembrane peptide, while FCGR3B is expressed on polymorphonuclear neutrophils (PMN) where the receptor is anchored through a phosphatidylinositol (PI) linkage. Mutations in this gene have been associated with immunodeficiency 20 and have been linked to susceptibility to recurrent viral infections, susceptibility to systemic lupus erythematosus, and alloimmune neonatal neutropenia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. Diseases associated with FCGR3A include Immunodeficiency 20 and Herpes Zoster.
            Bioinformatics
            Protein Aliases: CD16-I; CD16-II; CD16a; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III-A; Fc gamma receptor IIIa; Fc gamma receptor IIIb; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16); Fc-gamma receptor IIIb (CD16); Fc-gamma RII-b; Fc-gamma RII-c; Fc-gamma RIII; Fc-gamma RIII-alpha; Fc-gamma RIII-beta; Fc-gamma RIIIa; fc-gamma RIIIb; Fc-gamma-RIIb; Fc-gamma-RIIc; FCG2; FcgammaRIIIA; FCGR2; FCGR2A; FCGR2B; FCGR2C; FcR-10; FcRII-b; FcRII-c; IgG Fc receptor III-1; IgG Fc receptor III-2; IgG Fc receptor III-A; immunoglobulin G Fc receptor III; Low affinity immunoglobulin gamma Fc region receptor III-A; Low affinity immunoglobulin gamma Fc region receptor III-B; neutrophil-specific antigen NA; RP11-5K23.1
            View more View less
            Gene Aliases: CD16; CD16A; CD16B; FCG3; FCGR3; FCGR3A; FCGR3B; FCGRIII; FCR-10; FCRIII; FCRIIIA; FCRIIIb; IGFR3; IMD20
            View more View less
            UniProt ID: (Human) P08637, (Human) O75015
            View more View less
            Entrez Gene ID: (Human) 2214, (Human) 2215
            View more View less
            Function(s)
            IgG binding immunoglobulin receptor superfamily
            Process(es)
            immune response Fc-gamma receptor signaling pathway involved in phagocytosis regulation of immune response positive regulation of defense response to virus by host mitophagy in response to mitochondrial depolarization xenophagy
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-7c4bf66445-j9ktm:80/100.66.73.217:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0