Invitrogen antibodies—maximize lab budgets with advanced validation & coverage
Shop antibodies
    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Check Order Status
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • CD19 Antibodies
            Invitrogen
            CD19 Monoclonal Antibody (eBio1D3 (1D3)), Functional Grade, eBioscience™
            72 Published Figures
            134 References
            View all (351) CD19 antibodies
            Tech Support
            Cite CD19 Monoclonal Antibody (eBio1D3 (1D3)), Functional Grade, eBioscience™
            • Antibody Testing Data (1)
            • Published Figures (72)
            Application
            CD19 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.
            FIGURE: 1 / 73
            CD19 Antibody (16-0193-85) in Flow
            Staining of C57Bl/6 splenocytes with 0.25 µg of Rat IgG2a kappa Isotype Control Functional Grade Purified (Product # 16-4321-82) (open histogram) or 0.25 µg of Anti-Mouse CD19 Functional Grade Purified (filled histogram) followed by Anti-Rat IgG Biotin (Product # 13-4813-85) and Streptavidin PE (Product # 12-4317-87).Cells in the lymphocyte gate were used for analysis.
            View Product
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Immunohistochemistry (IHC)
            CD19 Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            CD19 Antibody in Flow Cytometry (Flow)
            Product Details
            16-0193-85
            Applications
            Tested Dilution
            Publications
            Immunohistochemistry (IHC)
            -
            View 4 publications 4 publications
            Immunohistochemistry (Frozen) (IHC (F))
            Assay-Dependent
            View 2 publications 2 publications
            Flow Cytometry (Flow)
            0.5 µg/test
            View 126 publications 126 publications
            Functional Assay (Functional)
            Assay-Dependent
            View 1 publication 1 publication
            Miscellaneous PubMed (Misc)
            -
            View 1 publication 1 publication
            Product Specifications
            Species Reactivity
            Mouse
            Published species
            Human, Mouse, Rat
            Host/Isotype
            Rat / IgG2a, kappa
            Recommended Isotype Control
            Rat IgG2a kappa Isotype Control (eBR2a), Functional Grade, eBioscience™
            Class
            Monoclonal
            Type
            Antibody
            Clone
            eBio1D3 (1D3)
            Conjugate
            Functional Grade
            • Unconjugated
            • Alexa Fluor 561
            • Alexa Fluor 660 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 496
            • Brilliant UV 563
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant UV 805
            • Brilliant Violet 421
            • Brilliant Violet 480
            • Brilliant Violet 605
            • Brilliant Violet 650
            • Brilliant Violet 711
            • Brilliant Violet 786
            • eFluor 450
            • eFluor 506
            • eFluor 660
            • FITC
            • NovaFluor Blue 510
            • NovaFluor Blue 585
            • NovaFluor Blue 610-70S
            • NovaFluor Blue 660-120S
            • NovaFluor Blue 690
            • NovaFluor Blue 725
            • NovaFluor Blue 760
            • NovaFluor Blue 800
            • NovaFluor Red 710
            • NovaFluor UV 765
            • NovaFluor Violet 690
            • NovaFluor Violet 800
            • NovaFluor Yellow 570
            • NovaFluor Yellow 590
            • NovaFluor Yellow 610
            • NovaFluor Yellow 660
            • NovaFluor Yellow 690
            • NovaFluor Yellow 700
            • NovaFluor Yellow 730
            • NovaFluor Yellow 755
            • NovaFluor Yellow 810
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-Cyanine7
            • PE-eFluor 610
            • PerCP-Cyanine5.5
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation
            Form
            Liquid
            Concentration
            1 mg/mL
            Purification
            Affinity chromatography
            Storage buffer
            PBS, pH 7.2
            Contains
            no preservative
            Storage conditions
            4°C
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            RRID
            AB_657670
            Product Specific Information
            Description: The eBio1D3 monoclonal antibody reacts with mouse CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, MHC class II, and CD19 form a multimolecular complex that associates with the BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells.
            Applications Reported: This eBio1D3 (1D3) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. The eBio1D3 monoclonal antibody has also been reported to induce down-regulation of CD19, affecting the proportions of B-1a and B-2 B cells in mice.
            Applications Tested: This eBio1D3 (1D3) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
            Storage and handling: Use in a sterile environment.
            Filtration: 0.2 µm post-manufacturing filtered.
            Purity: Greater than 90%, as determined by SDS-PAGE.
            Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
            Aggregation: Less than 10%, as determined by HPLC.
            Target Information
            CD19 is a member of the immunoglobulin superfamily, characterized by two Ig-like domains, and is expressed on B cells throughout all stages of development, excluding terminally differentiated plasma cells. It is also expressed on follicular dendritic cells and has been observed on myeloid leukemia cells, particularly those of monocytic lineage. CD19 is considered the earliest and broadest B cell-restricted antigen, and its expression is found in all B cell precursor leukemias. CD19 forms a multimolecular complex with CD21, CD81, Leu13, MHC class II, and the B cell receptor (BCR), playing a crucial role in B cell signaling. As a signal-amplifying coreceptor for the BCR, CD19 lowers the threshold for antigen receptor-dependent stimulation, allowing B cells to respond specifically and sensitively to various antigens through low-affinity antigen receptors. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. Beyond its role as a BCR coreceptor, CD19 can also signal independently of BCR co-ligation, serving as a central regulatory component upon which multiple signaling pathways converge. This makes CD19 an important functional regulator of both normal and malignant B cell proliferation. Mutations in the CD19 gene can result in hypogammaglobulinemia, a condition characterized by low levels of immunoglobulins, while CD19 overexpression can lead to B cell hyperactivity. CD19 is expressed on 100% of peripheral B cells, as defined by the expression of kappa or lambda light chains, underscoring its significance in B cell function and immune regulation.
            Bioinformatics
            Protein Aliases: B-lymphocyte antigen CD19; CD19; Differentiation antigen CD19; Leu-12
            View more View less
            Gene Aliases: AW495831; Cd19
            View more View less
            UniProt ID: (Mouse) P25918
            View more View less
            Entrez Gene ID: (Mouse) 12478
            View more View less
            Process(es)
            chromosome breakage B cell receptor signaling pathway positive regulation of release of sequestered calcium ion into cytosol
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            Spain flag icon
            Spain
            TEST
            Your items have has been added!
            Host server : magellan-search-green-56756dfb8b-7765t:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0