Product References
Extracellular HMGB-1 activates inflammatory signaling in tendon cells and tissues.
Therapeutic advances in chronic disease
Zhang C,Gu X,Zhao G,Wang W,Shao J,Zhu J,Yuan T,Sun J,Nie D,Zhou Y
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Sun Apr 17 00:00:00 UTC 2022
Inhibition of the JNK/MAPK signaling pathway by myogenesis-associated miRNAs is required for skeletal muscle development.
Cell death and differentiation
Xie SJ,Li JH,Chen HF,Tan YY,Liu SR,Zhang Y,Xu H,Yang JH,Liu S,Zheng LL,Huang MB,Guo YH,Zhang Q,Zhou H,Qu LH
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
Low-Dose Radiotherapy Has No Harmful Effects on Key Cells of Healthy Non-Inflamed Joints.
International journal of molecular sciences
Deloch L,Rückert M,Fietkau R,Frey B,Gaipl US
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Tue Oct 16 00:00:00 UTC 2018
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
The critical role of SENP1-mediated GATA2 deSUMOylation in promoting endothelial activation in graft arteriosclerosis.
Nature communications
Qiu C,Wang Y,Zhao H,Qin L,Shi Y,Zhu X,Song L,Zhou X,Chen J,Zhou H,Zhang H,Tellides G,Min W,Yu L
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2017
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Treg engage lymphotoxin beta receptor for afferent lymphatic transendothelial migration.
Nature communications
Brinkman CC,Iwami D,Hritzo MK,Xiong Y,Ahmad S,Simon T,Hippen KL,Blazar BR,Bromberg JS
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Tue Jun 21 00:00:00 UTC 2016
Distinct in vitro properties of embryonic and extraembryonic fibroblast-like cells are reflected in their in vivo behavior following grafting in the adult mouse brain.
Cell transplantation
Costa R,Bergwerf I,Santermans E,De Vocht N,Praet J,Daans J,Le Blon D,Hoornaert C,Reekmans K,Hens N,Goossens H,Berneman Z,Parolini O,Alviano F,Ponsaerts P
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Mon Nov 16 00:00:00 UTC 2015
Heme-mediated SPI-C induction promotes monocyte differentiation into iron-recycling macrophages.
Cell
Haldar M,Kohyama M,So AY,Kc W,Wu X,Briseño CG,Satpathy AT,Kretzer NM,Arase H,Rajasekaran NS,Wang L,Egawa T,Igarashi K,Baltimore D,Murphy TL,Murphy KM
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Flow Cytometry
Thu Mar 13 00:00:00 UTC 2014
Effects of intra-abdominal sepsis on atherosclerosis in mice.
Critical care (London, England)
Kaynar AM,Yende S,Zhu L,Frederick DR,Chambers R,Burton CL,Carter M,Stolz DB,Agostini B,Gregory AD,Nagarajan S,Shapiro SD,Angus DC
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Immunofluorescence
Wed Sep 03 00:00:00 UTC 2014
Nrf2 regulates haematopoietic stem cell function.
Nature cell biology
Tsai JJ,Dudakov JA,Takahashi K,Shieh JH,Velardi E,Holland AM,Singer NV,West ML,Smith OM,Young LF,Shono Y,Ghosh A,Hanash AM,Tran HT,Moore MA,van den Brink MR
16-1061 was used in Flow cytometry/Cell sorting to investigate the link between Nrf2 and CXCR4, thought to partially contribute to the maintenance of haematopoietic stem cell function.
Fri Mar 01 00:00:00 UTC 2013
Rhodamine-loaded intercellular adhesion molecule-1-targeted microbubbles for dual-modality imaging under controlled shear stresses.
Circulation. Cardiovascular imaging
Wu Z,Curaj A,Fokong S,Liehn EA,Weber C,Lammers T,Kiessling F,Zandvoort van M
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Immunohistochemistry
Fri Nov 01 00:00:00 UTC 2013
Rhodamine-loaded intercellular adhesion molecule-1-targeted microbubbles for dual-modality imaging under controlled shear stresses.
Circulation. Cardiovascular imaging
Wu Z,Curaj A,Fokong S,Liehn EA,Weber C,Lammers T,Kiessling F,Zandvoort van M
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Immunohistochemistry
Fri Nov 01 00:00:00 UTC 2013
Rhodamine-loaded intercellular adhesion molecule-1-targeted microbubbles for dual-modality imaging under controlled shear stresses.
Circulation. Cardiovascular imaging
Wu Z,Curaj A,Fokong S,Liehn EA,Weber C,Lammers T,Kiessling F,Zandvoort van M
Published figure using CD106 (VCAM-1) monoclonal antibody (Product # 16-1061-82) in Immunohistochemistry
Fri Nov 01 00:00:00 UTC 2013
Distinct sites within the vascular cell adhesion molecule-1 (VCAM-1) cytoplasmic domain regulate VCAM-1 activation of calcium fluxes versus Rac1 during leukocyte transendothelial migration.
Biochemistry
Marchese ME,Berdnikovs S,Cook-Mills JM
Tue Oct 16 00:00:00 UTC 2012