Product References
The Bisdioxopiperazine ICRF-193 Attenuates LPS-induced IL-1β Secretion by Macrophages.
Inflammation
Brindle A,Bainbridge C,Kumar MR,Todryk S,Padget K
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Myeloid‑derived suppressor cell accumulation induces Treg expansion and modulates lung malignancy progression.
Biomedical reports
Wan Y,Mu X,Zhao J,Li L,Xu W,Zhang M
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
ELMO1 Deficiency Reduces Neutrophil Chemotaxis in Murine Peritonitis.
International journal of molecular sciences
Yu S,Geng X,Liu H,Zhang Y,Cao X,Li B,Yan J
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sun Apr 30 00:00:00 UTC 2023
Regulation of synaptic connectivity in schizophrenia spectrum by mutual neuron-microglia interaction.
Communications biology
Breitmeyer R,Vogel S,Heider J,Hartmann SM,Wüst R,Keller AL,Binner A,Fitzgerald JC,Fallgatter AJ,Volkmer H
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sat Apr 29 00:00:00 UTC 2023
Local Myeloid-Derived Suppressor Cells Impair Progression of Experimental Autoimmune Uveitis by Alleviating Oxidative Stress and Inflammation.
Investigative ophthalmology & visual science
Lee JY,Kim S,Sohn HJ,Kim CH,Kim TG,Lee HS
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Oct 03 00:00:00 UTC 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2023
LAL deficiency induced myeloid-derived suppressor cells as targets and biomarkers for lung cancer.
Journal for immunotherapy of cancer
Zhao T,Liu S,Hanna NH,Jalal S,Ding X,Wan J,Yan C,Du H
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Methylation of SPRED1: A New Target in Acute Myeloid Leukemia.
Frontiers in oncology
Su N,Wang Y,Lu X,Xu W,Wang H,Mo W,Pang H,Tang R,Li S,Yan X,Li Y,Zhang R
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sat Apr 02 00:00:00 UTC 2022
Prognostic Lnc-S100B-2 Affects Cell Apoptosis and Microenvironment of Colorectal Cancer through MLLT10 Signaling.
Journal of oncology
Yi J,Peng F,Zhao J,Gong X
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Feb 08 00:00:00 UTC 2022
Pharmacological inhibition of LSD1 triggers myeloid differentiation by targeting GSE1 oncogenic functions in AML.
Oncogene
Nicosia L,Boffo FL,Ceccacci E,Conforti F,Pallavicini I,Bedin F,Ravasio R,Massignani E,Somervaille TCP,Minucci S,Bonaldi T
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Feb 01 00:00:00 UTC 2022
miR-204-containing exosomes ameliorate GVHD-associated dry eye disease.
Science advances
Zhou T,He C,Lai P,Yang Z,Liu Y,Xu H,Lin X,Ni B,Ju R,Yi W,Liang L,Pei D,Egwuagu CE,Liu X
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Fri Jan 14 00:00:00 UTC 2022
The Transcription of ZIP9 Is Associated With the Macrophage Polarization and the Pathogenesis of Hepatocellular Carcinoma.
Frontiers in immunology
Gou Y,Yang D,Tian T,Zhu X,Zhang R,Ren J,Tu D,Luo Y,Miao Y,Zhao H,Wang Y,Wei B
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Fri Apr 08 00:00:00 UTC 2022
Placenta-derived IL-32β activates neutrophils to promote preeclampsia development.
Cellular & molecular immunology
Liu D,Li Q,Ding H,Zhao G,Wang Z,Cao C,Dai Y,Zheng M,Zhu X,Wu Q,Wang Y,Duan H,Tang H,Lu X,Hou Y,Hu Y
17-0118-42 was used in Flow Cytometry to investigate whether IL-32 is expressed in the placenta and whether it contributes to the activation of neutrophils in preeclampsia patients.
Thu Apr 01 00:00:00 UTC 2021
Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
The Journal of experimental medicine
Louka E,Povinelli B,Rodriguez-Meira A,Buck G,Wen WX,Wang G,Sousos N,Ashley N,Hamblin A,Booth CAG,Roy A,Elliott N,Iskander D,de la Fuente J,Fordham N,O'Byrne S,Inglott S,Norfo R,Salio M,Thongjuea S,Rao A,Roberts I,Mead AJ
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Different Induction of PD-L1 (CD274) and PD-1 (CD279) Expression in THP-1-Differentiated Types 1 and 2 Macrophages.
Journal of inflammation research
Lai CY,Tseng PC,Chen CL,Satria RD,Wang YT,Lin CF
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Wed Apr 27 00:00:00 UTC 2022
G-MDSCs promote aging-related cardiac fibrosis by activating myofibroblasts and preventing senescence.
Cell death & disease
Sun SN,Ni SH,Li Y,Liu X,Deng JP,Chen ZX,Li H,Feng WJ,Huang YS,Li DN,Xian SX,Yang ZQ,Wang LJ,Lu L
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
From Active to Non-active Giant Cell Arteritis: Longitudinal Monitoring of Patients on Glucocorticoid Therapy in Combination With Leflunomide.
Frontiers in medicine
Kuret T,Frank-Bertoncelj M,Lakota K,Žigon P,Thallinger GG,Kopitar AN,Čučnik S,Tomšič M,Hočevar A,Sodin-Šemrl S
17-0118-42 was used in Flow cytometry/Cell sorting to monitor leukocyte subsets, expression of neutrophil surface adhesion molecules (CD62L and CD11b) and serum analytes in therapy-naive patients with active giant cell arteritis (GCA).
Tue Feb 08 00:00:00 UTC 2022
Associations between Hypertriglyceridemia and Circulating Neutrophil Subpopulation in Patients with Dyslipidemia.
International journal of inflammation
Genkel V,Dolgushin I,Baturina I,Savochkina A,Kuznetsova A,Pykhova L,Shaposhnik I
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sun Apr 24 00:00:00 UTC 2022
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
17-0118 was used in Flow cytometry/Cell sorting to determine the basis for aberrant megakaryopoiesis in myelofibrosis using single-cell-omics.
Thu May 07 00:00:00 UTC 2020
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
17-0118 was used in Flow cytometry/Cell sorting to determine the basis for aberrant megakaryopoiesis in myelofibrosis using single-cell-omics.
Thu May 07 00:00:00 UTC 2020
The long non-coding RNA Cancer Susceptibility 15 (CASC15) is induced by isocitrate dehydrogenase (IDH) mutations and maintains an immature phenotype in adult acute myeloid leukemia.
Haematologica
Grasedieck S,Ruess C,Krowiorz K,Lux S,Pochert N,Schwarzer A,Klusmann JH,Jongen-Lavrencic M,Herold T,Bullinger L,Pollack JR,Rouhi A,Kuchenbauer F
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
The long non-coding RNA Cancer Susceptibility 15 (CASC15) is induced by isocitrate dehydrogenase (IDH) mutations and maintains an immature phenotype in adult acute myeloid leukemia.
Haematologica
Grasedieck S,Ruess C,Krowiorz K,Lux S,Pochert N,Schwarzer A,Klusmann JH,Jongen-Lavrencic M,Herold T,Bullinger L,Pollack JR,Rouhi A,Kuchenbauer F
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Galectin-9 promotes a suppressive microenvironment in human cancer by enhancing STING degradation.
Oncogenesis
Zhang CX,Huang DJ,Baloche V,Zhang L,Xu JX,Li BW,Zhao XR,He J,Mai HQ,Chen QY,Zhang XS,Busson P,Cui J,Li J
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Mon Jul 06 00:00:00 UTC 2020
Connecting METTL3 and intratumoural CD33+ MDSCs in predicting clinical outcome in cervical cancer.
Journal of translational medicine
Ni HH,Zhang L,Huang H,Dai SQ,Li J
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Oct 15 00:00:00 UTC 2020
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Wed Feb 19 00:00:00 UTC 2020
circASXL1-1 regulates BAP1 deubiquitinase activity in leukemia.
Haematologica
Jadhav SP,Kumari N,Ng L,Tan PF,Yeo-Teh NSL,Goh Y,Fam WN,Tng JQ,Tian JS,Koh BTH,Chun CM,Wang W,Chng WJ,Fullwood MJ,Guccione E,Karnani N,Tenen DG,Jha S
17-0118-42 was used in Flow Cytometry to show a new mechanism for regulation of H2AK119ub levels: via interaction of circASXL1-1 and BAP1.
Wed Jul 01 00:00:00 UTC 2020
Overexpression of ERBB4 rejuvenates aged mesenchymal stem cells and enhances angiogenesis via PI3K/AKT and MAPK/ERK pathways.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liang X,Ding Y,Lin F,Zhang Y,Zhou X,Meng Q,Lu X,Jiang G,Zhu H,Chen Y,Lian Q,Fan H,Liu Z
17-0118-42 was used in Flow Cytometry to investigate whether Erb-B2 receptor tyrosine kinase 4 ERBB4 rejuvenates aged mesenchymal stem cells MSC and how ERBB4 enhances therapeutic efficacy of aged MSC in treating myocardial infarction (MI).
Fri Mar 01 00:00:00 UTC 2019
Ubiquitin-specific peptidase 3 induces TPA-mediated leukemia cell differentiation via regulating H2AK119ub.
Animal cells and systems
Chae YC,Jung H,Kim JY,Lee DH,Seo SB
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Sep 29 00:00:00 UTC 2020
Microbiome Influences Prenatal and Adult Microglia in a Sex-Specific Manner.
Cell
Thion MS,Low D,Silvin A,Chen J,Grisel P,Schulte-Schrepping J,Blecher R,Ulas T,Squarzoni P,Hoeffel G,Coulpier F,Siopi E,David FS,Scholz C,Shihui F,Lum J,Amoyo AA,Larbi A,Poidinger M,Buttgereit A,Lledo PM,Greter M,Chan JKY,Amit I,Beyer M,Schultze JL,Schlitzer A,Pettersson S,Ginhoux F,Garel S
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Jan 25 00:00:00 UTC 2018
AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
Molecules and cells
Park JW,Cho H,Oh H,Kim JY,Seo SB
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu May 31 00:00:00 UTC 2018
n-butanol extract from Folium isatidis inhibits the lipopolysaccharide-induced downregulation of CXCR1 and CXCR2 on human neutrophils.
Molecular medicine reports
Wu B,Wang L,Jiang L,Dong L,Xu F,Lu Y,Jin J,Wang Z,Liang G,Shan X
Mon Jan 01 00:00:00 UTC 2018
Accumulation of T-helper 22 cells, interleukin-22 and myeloid-derived suppressor cells promotes gastric cancer progression in elderly patients.
Oncology letters
Chen X,Wang Y,Wang J,Wen J,Jia X,Wang X,Zhang H
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sun Jul 01 00:00:00 UTC 2018
Deacetylase activity-independent transcriptional activation by HDAC2 during TPA-induced HL-60 cell differentiation.
PloS one
Jung H,Kim JY,Kim KB,Chae YC,Hahn Y,Kim JW,Seo SB
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Feb 07 00:00:00 UTC 2019
Microglia innately develop within cerebral organoids.
Nature communications
Ormel PR,Vieira de Sá R,van Bodegraven EJ,Karst H,Harschnitz O,Sneeboer MAM,Johansen LE,van Dijk RE,Scheefhals N,Berdenis van Berlekom A,Ribes Martínez E,Kling S,MacGillavry HD,van den Berg LH,Kahn RS,Hol EM,de Witte LD,Pasterkamp RJ
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Oct 09 00:00:00 UTC 2018
Human-induced pluripotent stem cell-derived macrophages and their immunological function in response to tuberculosis infection.
Stem cell research & therapy
Hong D,Ding J,Li O,He Q,Ke M,Zhu M,Liu L,Ou WB,He Y,Wu Y
17-0118 was used in Flow cytometry/Cell sorting to characterise human induced pluripotent stem cell-derived macrophages and their response to Bacillus Calmette-Guerin infection.
Mon Feb 26 00:00:00 UTC 2018
Bioreactor-based mass production of human iPSC-derived macrophages enables immunotherapies against bacterial airway infections.
Nature communications
Ackermann M,Kempf H,Hetzel M,Hesse C,Hashtchin AR,Brinkert K,Schott JW,Haake K,Kühnel MP,Glage S,Figueiredo C,Jonigk D,Sewald K,Schambach A,Wronski S,Moritz T,Martin U,Zweigerdt R,Munder A,Lachmann N
17-0118 was used in Flow cytometry/Cell sorting to develop a novel, cell-based treatment strategy to target bacterial airway infections.
Fri Nov 30 00:00:00 UTC 2018
The prognostic significance of CD11b+CX3CR1+ monocytes in patients with newly diagnosed diffuse large B-cell lymphoma.
Oncotarget
Yhim HY,Kim JA,Ko SH,Park Y,Yim E,Kim HS,Kwak JY
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Fri Nov 03 00:00:00 UTC 2017
KSRP specifies monocytic and granulocytic differentiation through regulating miR-129 biogenesis and RUNX1 expression.
Nature communications
Zhao H,Wang X,Yi P,Si Y,Tan P,He J,Yu S,Ren Y,Ma Y,Zhang J,Wang D,Wang F,Yu J
17-0118 was used in Flow cytometry/Cell sorting to elucidate how KH-Type Splicing Regulatory Protein, miR-129, and RUNX1 participate in a regulatory axis to control the outcome of myeloid differentiation.
Fri Nov 10 00:00:00 UTC 2017
TLR2 activation induces antioxidant defence in human monocyte-macrophage cell line models.
Oncotarget
Karwaciak I,Gorzkiewicz M,Bartosz G,Pulaski L
17-0118 was used in Flow cytometry/Cell sorting to determine whether there are specific biochemical mechanisms responsible for an increase in oxidative stress resistance in differentiating macrophages.
Tue Aug 15 00:00:00 UTC 2017
CD19 CAR immune pressure induces B-precursor acute lymphoblastic leukaemia lineage switch exposing inherent leukaemic plasticity.
Nature communications
Jacoby E,Nguyen SM,Fountaine TJ,Welp K,Gryder B,Qin H,Yang Y,Chien CD,Seif AE,Lei H,Song YK,Khan J,Lee DW,Mackall CL,Gardner RA,Jensen MC,Shern JF,Fry TJ
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Wed Jul 27 00:00:00 UTC 2016
Vitamin D machinery and metabolism in porcine adipose-derived mesenchymal stem cells.
Stem cell research & therapy
Valle YL,Almalki SG,Agrawal DK
17-0118 was used in Flow cytometry/Cell sorting to evaluate whether porcine adipose-derived MSCs can metabolise and respond to vitamin D.
Wed Aug 17 00:00:00 UTC 2016
Metabolic regulation of hepatitis B immunopathology by myeloid-derived suppressor cells.
Nature medicine
Pallett LJ,Gill US,Quaglia A,Sinclair LV,Jover-Cobos M,Schurich A,Singh KP,Thomas N,Das A,Chen A,Fusai G,Bertoletti A,Cantrell DA,Kennedy PT,Davies NA,Haniffa M,Maini MK
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
STK4 regulates TLR pathways and protects against chronic inflammation-related hepatocellular carcinoma.
The Journal of clinical investigation
Li W,Xiao J,Zhou X,Xu M,Hu C,Xu X,Lu Y,Liu C,Xue S,Nie L,Zhang H,Li Z,Zhang Y,Ji F,Hui L,Tao W,Wei B,Wang H
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Mon Nov 02 00:00:00 UTC 2015
Affinity maturation of a novel antagonistic human monoclonal antibody with a long VH CDR3 targeting the Class A GPCR formyl-peptide receptor 1.
mAbs
Douthwaite JA,Sridharan S,Huntington C,Hammersley J,Marwood R,Hakulinen JK,Ek M,Sjögren T,Rider D,Privezentzev C,Seaman JC,Cariuk P,Knights V,Young J,Wilkinson T,Sleeman M,Finch DK,Lowe DC,Vaughan TJ
17-0118 was used in Flow cytometry/Cell sorting to illustrate an approach for antibody discovery and affinity engineering to typically intractable membrane proteins such as GPCRs.
Mon Sep 28 00:00:00 UTC 2015
Induction of autophagy is a key component of all-trans-retinoic acid-induced differentiation in leukemia cells and a potential target for pharmacologic modulation.
Experimental hematology
Orfali N,O'Donovan TR,Nyhan MJ,Britschgi A,Tschan MP,Cahill MR,Mongan NP,Gudas LJ,McKenna SL
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
dNP2 is a blood-brain barrier-permeable peptide enabling ctCTLA-4 protein delivery to ameliorate experimental autoimmune encephalomyelitis.
Nature communications
Lim S,Kim WJ,Kim YH,Lee S,Koo JH,Lee JA,Yoon H,Kim DH,Park HJ,Kim HM,Lee HG,Yun Kim J,Lee JU,Hun Shin J,Kyun Kim L,Doh J,Kim H,Lee SK,Bothwell ALM,Suh M,Choi JM
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Tue Sep 15 00:00:00 UTC 2015
Pre-Vaccination Frequencies of Th17 Cells Correlate with Vaccine-Induced T-Cell Responses to Survivin-Derived Peptide Epitopes.
PloS one
Køllgaard T,Ugurel-Becker S,Idorn M,Andersen MH,Becker JC,Straten PT
17-0118 was used in Flow cytometry/Cell sorting to investigate correlations between Th17 cells, MDSCs and Tregs and vaccine-induced T-cell responses, showing that pre-vaccination frequencies of Tregs correlate with responses to survivin-derived peptide epitopes.
Fri Apr 29 00:00:00 UTC 2016
A breast cancer stem cell niche supported by juxtacrine signalling from monocytes and macrophages.
Nature cell biology
Lu H,Clauser KR,Tam WL,Fröse J,Ye X,Eaton EN,Reinhardt F,Donnenberg VS,Bhargava R,Carr SA,Weinberg RA
17-0118 was used in Flow cytometry/Cell sorting to investigate the interactions between cancer stem cells and their surrounding microenvironment.
Sat Nov 01 00:00:00 UTC 2014
Mast cell histamine promotes the immunoregulatory activity of myeloid-derived suppressor cells.
Journal of leukocyte biology
Martin RK,Saleem SJ,Folgosa L,Zellner HB,Damle SR,Nguyen GK,Ryan JJ,Bear HD,Irani AM,Conrad DH
17-0118 was used in Flow cytometry/Cell sorting to indicate that MCs and MC-released histamine are critical for MDSC-mediated immune regulation.
Tue Jul 01 00:00:00 UTC 2014
A method for high purity intestinal epithelial cell culture from adult human and murine tissues for the investigation of innate immune function.
Journal of immunological methods
Graves CL,Harden SW,LaPato M,Nelson M,Amador B,Sorenson H,Frazier CJ,Wallet SM
17-0118 was used in Flow cytometry/Cell sorting to demonstrate a novel culture method for more appropriate study of intestinal epithelial cells as innate immune effectors.
Mon Dec 01 00:00:00 UTC 2014
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) preserves gut barrier function by blocking neutrophil-endothelial cell adhesion after hemorrhagic shock and resuscitation in mice.
Surgery
Zhang HY,James I,Chen CL,Besner GE
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2012
Normal human monocytes exposed to glioma cells acquire myeloid-derived suppressor cell-like properties.
Neuro-oncology
Rodrigues JC,Gonzalez GC,Zhang L,Ibrahim G,Kelly JJ,Gustafson MP,Lin Y,Dietz AB,Forsyth PA,Yong VW,Parney IF
Published figure using CD11b monoclonal antibody (Product # 17-0118-42) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2010
Flotillin microdomains interact with the cortical cytoskeleton to control uropod formation and neutrophil recruitment.
The Journal of cell biology
Ludwig A,Otto GP,Riento K,Hams E,Fallon PG,Nichols BJ
17-0118 was used in Flow cytometry/Cell sorting to study the role of flotillin microdomain-cortical cytoskeleton associations in neutrophil migration, urpopod formation, and regulation of myosin IIa.
Mon Nov 15 00:00:00 UTC 2010