Invitrogen antibodies—maximize lab budgets with advanced validation & coverage
Shop antibodies
    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • SIRP alpha (CD172a) Antibodies
            The product (cat # 25-1721-80) that you're looking for has been discontinued and replaced with 25-1721-82. The product (cat # 25-1721-80) that you're looking for has been discontinued. Please contact Technical Support with any questions.
            Invitrogen
            CD172a (SIRP alpha) Monoclonal Antibody (P84), PE-Cyanine7, eBioscience™
            1 Published Figure
            2 References
            View all (82) SIRP alpha (CD172a) antibodies
            Datasheet
            Protocols
            Catalog # 25-1721-80
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
            This product has been discontinued
            25 µg
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Cite CD172a (SIRP alpha) Monoclonal Antibody (P84), PE-Cyanine7, eBioscience™
            • Antibody Testing Data (1)
            • Published Figures (1)
            Application
            CD172a (SIRP alpha) Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.
            FIGURE: 1 / 2
            CD172a (SIRP alpha) Antibody (25-1721-80) in Flow
            Swiss Webster mouse bone marrow cells were stained with CD11b Monoclonal Antibody, FITC (Product # 11-0112-82) and 0.5 µg of Rat IgG1 kappa Isotype Control, PE-Cyanine7 (Product # 25-4301-82) (left) or 0.5 µg of CD172a Monoclonal Antibody, PE-Cyanine7 (right). Total viable cells were used for analysis.
            View Product
            CD172a (SIRP alpha) Antibody in Flow Cytometry (Flow)
            CD172a (SIRP alpha) Antibody in Flow Cytometry (Flow)
            Product Details
            25-1721-80
            Applications
            Tested Dilution
            Publications
            Flow Cytometry (Flow)
            1 µg/test
            View 2 publications 2 publications
            Product Specifications
            Host/Isotype
            Rat / IgG1, kappa
            Recommended Isotype Control
            Rat IgG1 kappa Isotype Control (eBRG1), PE-Cyanine7, eBioscience™
            Class
            Monoclonal
            Type
            Antibody
            Clone
            P84
            Conjugate
            PE-Cyanine7
            • Alexa Fluor 488 
            • APC 
            • Biotin 
            • Functional Grade
            • PE 
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 645
            • Request custom conjugation
            Excitation/Emission Max
            569/780 nm View spectra spectra
            Form
            Liquid
            Concentration
            0.2 mg/mL
            Storage conditions
            4°C, store in dark, DO NOT FREEZE!
            Shipping conditions
            Wet ice
            RRID
            AB_2762639
            Product Specific Information
            Description: This P84 monoclonal antibody reacts with mouse CD172a, also known as signal regulatory protein a (SIRPa). This cell surface glycoprotein consists of three Ig-like extracellular domains and two cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The ITIM domains have been demonstrated to recruit and bind the Src homology 2 domain-containing phosphatases SHP-1 and SHP-2. CD172a is expressed on monocytes, macrophages, dendritic cells, but not on T and B lymphocytes. Moreover, neurons and other tissues of the central nervous system have also been shown to express CD172a. The integrin-associated protein CD47 is the extracellular ligand for CD172a. Studies show that CD172a is involved in dendritic cell-mediated T cell activation, neutrophil migration, and phagocytosis.
            This monoclonal antibody has been reported to have neutralizing activity.
            Applications Reported: This P84 antibody has been reported for use in flow cytometric analysis.
            Applications Tested: This P84 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 1.0 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
            Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
            Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
            Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
            Target Information
            SIRP alpha (CD172a, signal-regulatory protein alpha) is a receptor-type transmembrane glycoprotein expressed on cells of myeloid origin, including granulocytes, dendritic cells (DCs), macrophages, mast cells and hematopoietic stem cells. SIRP alpha acts as a substrate for several activated tyrosine kinases, including EGFR, PDGFR, src and insulin receptor and is involved in the negative regulation of receptor tyrosine kinase-coupled signaling pathways. The ligand binding of SIRP alpha to integrin-associated protein CD47 results in tyrosine kinase phosphorylation of immunoreceptor tyrosine-based inhibitory motifs (ITIMs) within the cytoplasmic region of SIRP alpha, which mediates the recruitment and activation of the tyrosine phosphatases SHP-1 and SHP-2. Ligation of SIRP alpha with CD47 has been demonstrated in several regulatory processes, including the inhibition of host cell phagocytosis by macrophages and the bi-directional activation of T cells and DCs. SIRP alpha has regulatory effects on cellular responses induced by serum, growth factors, insulin, oncogenes, growth hormones and cell adhesion, and plays a general role in different physiological and pathological processes. Cancer cells highly express CD47, which activates SIRP alpha and inhibits macrophage-mediated destruction of cancerous cell growth.
            How to use the Panel Builder
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-7c4bf66445-zkq99:80/100.66.72.19:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0