Product References
miR-539-5p targets BMP2 to regulate Treg activation in B-cell acute lymphoblastic leukemia through TGF-β/Smads/MAPK.
Experimental biology and medicine (Maywood, N.J.)
Dai Q,Shi R,Zhang G,Wang Y,Ye L,Peng L,Guo S,He J,Yang H,Jiang Y
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Indole-3-pyruvic acid alleviates rheumatoid arthritis via the aryl hydrocarbon receptor pathway.
Annals of translational medicine
Huang T,Cheng L,Jiang Y,Zhang L,Qian L
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Wed Mar 15 00:00:00 UTC 2023
Advanced immunophenotyping: A powerful tool for immune profiling, drug screening, and a personalized treatment approach.
Frontiers in immunology
Preglej T,Brinkmann M,Steiner G,Aletaha D,Göschl L,Bonelli M
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Apr 11 00:00:00 UTC 2023
Rotenone aggravates PD-like pathology in A53T mutant human α-synuclein transgenic mice in an age-dependent manner.
Frontiers in aging neuroscience
Chen AD,Cao JX,Chen HC,Du HL,Xi XX,Sun J,Yin J,Jing YH,Gao LP
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Fri Aug 26 00:00:00 UTC 2022
Notch4 signaling limits regulatory T-cell-mediated tissue repair and promotes severe lung inflammation in viral infections.
Immunity
Harb H,Benamar M,Lai PS,Contini P,Griffith JW,Crestani E,Schmitz-Abe K,Chen Q,Fong J,Marri L,Filaci G,Del Zotto G,Pishesha N,Kolifrath S,Broggi A,Ghosh S,Gelmez MY,Oktelik FB,Cetin EA,Kiykim A,Kose M,Wang Z,Cui Y,Yu XG,Li JZ,Berra L,Stephen-Victor E,Charbonnier LM,Zanoni I,Ploegh H,Deniz G,De Palma R,Chatila TA
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Jun 08 00:00:00 UTC 2021
FABP4 facilitates inflammasome activation to induce the Treg/Th17 imbalance in preeclampsia via forming a positive feedback with IL-17A.
Molecular therapy. Nucleic acids
Chang GP,Yang XL,Liu W,Lin S,Yang SL,Zhao MY
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Fri Jun 04 00:00:00 UTC 2021
Soluble fibrinogen‑like protein 2 levels are decreased in patients with ischemic heart failure and associated with cardiac function.
Molecular medicine reports
You Y,Huang S,Liu H,Fan C,Liu K,Wang Z
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
T cell receptor-dependent S-acylation of ZAP-70 controls activation of T cells.
The Journal of biological chemistry
Tewari R,Shayahati B,Fan Y,Akimzhanov AM
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Thu Aug 26 00:00:00 UTC 2021
Identification of celastrol as a novel HIV-1 latency reversal agent by an image-based screen.
PloS one
Liu H,Hu PW,Dubrulle J,Stossi F,Nikolai BC,Mancini MA,Rice AP
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Sep 14 00:00:00 UTC 2021
Numerical optimization of microfluidic vortex shedding for genome editing T cells with Cas9.
Scientific reports
Jarrell JA,Sytsma BJ,Wilson LH,Pan FL,Lau KHWJ,Kirby GTS,Lievano AA,Pawell RS
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Thu Jun 03 00:00:00 UTC 2021
Human endoglin-CD3 bispecific T cell engager antibody induces anti-tumor effect in vivo.
Theranostics
Zhong L,Shi W,Gan L,Liu X,Huo Y,Wu P,Zhang Z,Wu T,Peng H,Huang Y,Zhao Y,Yuan Y,Deng Z,Tang H
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Wed Jul 28 00:00:00 UTC 2021
Epinephrine Production in Th17 Cells and Experimental Autoimmune Encephalitis.
Frontiers in immunology
Yang P,Tian H,Zou YR,Chambon P,Ichinose H,Honig G,Diamond B,Kim SJ
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Mon Jul 05 00:00:00 UTC 2021
Potential of neutrophil to lymphocyte ratio in predicting sustained remission in rheumatoid arthritis compared to other immune activation markers.
The Indian journal of medical research
Chandrashekara S,Lingaraju DC,Renuka P,Anupama KR
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2020
Involvement of natural killer cells in the pathogenesis of endometriosis in patients with pelvic pain.
The Journal of international medical research
He J,Xu Y,Yi M,Gu C,Zhu Y,Hu G
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
Th17 reprogramming of T cells in systemic juvenile idiopathic arthritis.
JCI insight
Henderson LA,Hoyt KJ,Lee PY,Rao DA,Jonsson AH,Nguyen JP,Rutherford K,Julé AM,Charbonnier LM,Case S,Chang MH,Cohen EM,Dedeoglu F,Fuhlbrigge RC,Halyabar O,Hazen MM,Janssen E,Kim S,Lo J,Lo MS,Meidan E,Son MBF,Sundel RP,Stoll ML,Nusbaum C,Lederer JA,Chatila TA,Nigrovic PA
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Thu Mar 26 00:00:00 UTC 2020
Th2 Biased Immunity With Altered B Cell Profiles in Circulation of Patients With Sporotrichosis Caused by Sporothrix globosa.
Frontiers in immunology
Zu J,Yao L,Song Y,Cui Y,Guan M,Chen R,Zhen Y,Li S
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Mon May 31 00:00:00 UTC 2021
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2020
Tumors establish resistance to immunotherapy by regulating Treg recruitment via CCR4.
Journal for immunotherapy of cancer
Marshall LA,Marubayashi S,Jorapur A,Jacobson S,Zibinsky M,Robles O,Hu DX,Jackson JJ,Pookot D,Sanchez J,Brovarney M,Wadsworth A,Chian D,Wustrow D,Kassner PD,Cutler G,Wong B,Brockstedt DG,Talay O
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Loading of Primary Human T Lymphocytes with Citrate-Coated Superparamagnetic Iron Oxide Nanoparticles Does Not Impair Their Activation after Polyclonal Stimulation.
Cells
Mühlberger M,Unterweger H,Band J,Lehmann C,Heger L,Dudziak D,Alexiou C,Lee G,Janko C
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Transcriptional down-regulation of ccr5 in a subset of HIV+ controllers and their family members.
eLife
Gonzalo-Gil E,Rapuano PB,Ikediobi U,Leibowitz R,Mehta S,Coskun AK,Porterfield JZ,Lampkin TD,Marconi VC,Rimland D,Walker BD,Deeks S,Sutton RE
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Apr 09 00:00:00 UTC 2019
Alteration of Th17 and Foxp3+ regulatory T cells in patients with unexplained recurrent spontaneous abortion before and after the therapy of hCG combined with immunoglobulin.
Experimental and therapeutic medicine
Sha J,Liu F,Zhai J,Liu X,Zhang Q,Zhang B
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD25 monoclonal antibody (Product # 46-0259-42) in Flow Cytometry
Fri Dec 15 00:00:00 UTC 2017