Product References
Bystander activated CD8+ T cells mediate neuropathology during viral infection via antigen-independent cytotoxicity.
Nature communications
Balint E,Feng E,Giles EC,Ritchie TM,Qian AS,Vahedi F,Montemarano A,Portillo AL,Monteiro JK,Trigatti BL,Ashkar AA
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2024
Treatment with glatiramer acetate in APPswe/PS1dE9 mice at an early stage of Alzheimer's disease prior to amyloid-beta deposition delays the disease's pathological development and ameliorates cognitive decline.
Frontiers in aging neuroscience
Huang Z,Gong Z,Lin Y,Yang F,Chen W,Xiang S,Huang Y,Xiao H,Xu S,Duan J
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Thu Feb 15 00:00:00 UTC 2024
6-Mercaptopurine potently inhibits recruitment of SHP2 by phosphorylated PD-1 to inhibit PD-1 signalling and enhance T cell function.
Immunology
Liu L,Lei Y,Zheng Z,Zhou X,Chen S,Zeng G,Yu L,Wang P,Chen L
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2023
An essential role for miR-15/16 in Treg suppression and restriction of proliferation.
Cell reports
Johansson K,Gagnon JD,Zhou SK,Fassett MS,Schroeder AW,Kageyama R,Bautista RA,Pham H,Woodruff PG,Ansel KM
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Oct 31 00:00:00 UTC 2023
Local Myeloid-Derived Suppressor Cells Impair Progression of Experimental Autoimmune Uveitis by Alleviating Oxidative Stress and Inflammation.
Investigative ophthalmology & visual science
Lee JY,Kim S,Sohn HJ,Kim CH,Kim TG,Lee HS
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Oct 03 00:00:00 UTC 2023
Amyloid-β specific regulatory T cells attenuate Alzheimer's disease pathobiology in APP/PS1 mice.
Molecular neurodegeneration
Yeapuri P,Machhi J,Lu Y,Abdelmoaty MM,Kadry R,Patel M,Bhattarai S,Lu E,Namminga KL,Olson KE,Foster EG,Mosley RL,Gendelman HE
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Dec 18 00:00:00 UTC 2023
CD138 promotes the accumulation and activation of autoreactive T cells in autoimmune MRL/lpr mice.
Experimental and therapeutic medicine
Xie T,Liu X,Li P
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
Regulatory T-cells and IL-5 mediate pain outcomes in a preclinical model of chronic muscle pain.
Molecular pain
Lenert ME,Szabo-Pardi TA,Burton MD
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Feb 13 00:00:00 UTC 2023
Immunotherapy targeting plasma ASM is protective in a mouse model of Alzheimer's disease.
Nature communications
Choi BJ,Park MH,Park KH,Han WH,Yoon HJ,Jung HY,Hong JY,Chowdhury MR,Kim KY,Lee J,Song IS,Pang M,Choi MK,Gulbins E,Reichel M,Kornhuber J,Hong CW,Kim C,Kim SH,Schuchman EH,Jin HK,Bae JS
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Mar 24 00:00:00 UTC 2023
Group 2 Innate Lymphoid Cells Protect Mice from Abdominal Aortic Aneurysm Formation via IL5 and Eosinophils.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zhang Y,Liu T,Deng Z,Fang W,Zhang X,Zhang S,Wang M,Luo S,Meng Z,Liu J,Sukhova GK,Li D,McKenzie ANJ,Libby P,Shi GP,Guo J
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2023
Aiolos represses CD4+ T cell cytotoxic programming via reciprocal regulation of TFH transcription factors and IL-2 sensitivity.
Nature communications
Read KA,Jones DM,Pokhrel S,Hales EDS,Varkey A,Tuazon JA,Eisele CD,Abdouni O,Saadey A,Leonard MR,Warren RT,Powell MD,Boss JM,Hemann EA,Yount JS,Xin G,Ghoneim HE,Lio CJ,Freud AG,Collins PL,Oestreich KJ
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Mar 24 00:00:00 UTC 2023
A hydrogen peroxide economizer for on-demand oxygen production-assisted robust sonodynamic immunotherapy.
Theranostics
Jiang Q,Qiao B,Lin X,Cao J,Zhang N,Guo H,Liu W,Zhu L,Xie X,Wan L,Tang R,Liang B,Wang D,Wang Z,Zhou Y,Ran H,Li P
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2022
Glutamine is essential for overcoming the immunosuppressive microenvironment in malignant salivary gland tumors.
Theranostics
Cao S,Hung YW,Wang YC,Chung Y,Qi Y,Ouyang C,Zhong X,Hu W,Coblentz A,Maghami E,Sun Z,Lin HH,Ann DK
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Aug 16 00:00:00 UTC 2022
Stage-specific action of Runx1 and GATA3 controls silencing of PU.1 expression in mouse pro-T cells.
The Journal of experimental medicine
Hosokawa H,Koizumi M,Masuhara K,Romero-Wolf M,Tanaka T,Nakayama T,Rothenberg EV
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Aug 02 00:00:00 UTC 2021
Allergen-Specific Treg Cells Upregulated by Lung-Stage S. japonicum Infection Alleviates Allergic Airway Inflammation.
Frontiers in cell and developmental biology
Li Z,Zhang W,Luo F,Li J,Yang W,Zhu B,Wu Q,Wang X,Sun C,Xie Y,Xu B,Wang Z,Qian F,Chen J,Wan Y,Hu W
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Neutralization
Sat Jun 26 00:00:00 UTC 2021
TRPM8 channel augments T-cell activation and proliferation.
Cell biology international
Acharya TK,Tiwari A,Majhi RK,Goswami C
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
MicroRNA‑21‑5p protects melanocytes via targeting STAT3 and modulating Treg/Teff balance to alleviate vitiligo.
Molecular medicine reports
Huo J,Liu T,Li F,Song X,Hou X
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Irradiated whole-cell vaccine suppresses hepatocellular carcinoma growth in mice via Th9 cells.
Oncology letters
Chen J,Ding Y,Huang F,Lan R,Wang Z,Huang W,Chen R,Wu B,Fu L,Yang Y,Liu J,Hong J,Zhang W,Zhang L
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Sat May 01 00:00:00 UTC 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Cell-permeable transgelin-2 as a potent therapeutic for dendritic cell-based cancer immunotherapy.
Journal of hematology & oncology
Kim HR,Park JS,Park JH,Yasmin F,Kim CH,Oh SK,Chung IJ,Jun CD
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Mar 17 00:00:00 UTC 2021
Neuroimmune Consequences of eIF4E Phosphorylation on Chemotherapy-Induced Peripheral Neuropathy.
Frontiers in immunology
Agalave NM,Mody PH,Szabo-Pardi TA,Jeong HS,Burton MD
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Sep 20 00:00:00 UTC 2021
Positive Effects of Oral Antibiotic Administration in Murine Chronic Graft-Versus-Host Disease.
International journal of molecular sciences
Sato S,Shimizu E,He J,Ogawa M,Asai K,Yazu H,Rusch R,Yamane M,Yang F,Fukuda S,Kawakami Y,Tsubota K,Ogawa Y
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Immunohistochemistry
Sat Apr 03 00:00:00 UTC 2021
Aiduqing formula inhibits breast cancer metastasis by suppressing TAM/CXCL1-induced Treg differentiation and infiltration.
Cell communication and signaling : CCS
Li J,Wang S,Wang N,Zheng Y,Yang B,Wang X,Zhang J,Pan B,Wang Z
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Aug 30 00:00:00 UTC 2021
The effect of fingolimod on regulatory T cells in a mouse model of brain ischaemia.
Journal of neuroinflammation
Malone K,Diaz Diaz AC,Shearer JA,Moore AC,Waeber C
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Sat Jan 30 00:00:00 UTC 2021
PI3Kδ coordinates transcriptional, chromatin, and metabolic changes to promote effector CD8+ T cells at the expense of central memory.
Cell reports
Cannons JL,Villarino AV,Kapnick SM,Preite S,Shih HY,Gomez-Rodriguez J,Kaul Z,Shibata H,Reilley JM,Huang B,Handon R,McBain IT,Gossa S,Wu T,Su HC,McGavern DB,O'Shea JJ,McGuire PJ,Uzel G,Schwartzberg PL
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Oct 12 00:00:00 UTC 2021
Regulatory T cells function in established systemic inflammation and reverse fatal autoimmunity.
Nature immunology
Hu W,Wang ZM,Feng Y,Schizas M,Hoyos BE,van der Veeken J,Verter JG,Bou-Puerto R,Rudensky AY
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Sep 01 00:00:00 UTC 2021
Acacetin ameliorates insulin resistance in obesity mice through regulating Treg/Th17 balance via MiR-23b-3p/NEU1 Axis.
BMC endocrine disorders
Wei Y,Jing J,Peng Z,Liu X,Wang X
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Mar 29 00:00:00 UTC 2021
Near-infrared photoimmunotherapy targeting human-EGFR in a mouse tumor model simulating current and future clinical trials.
EBioMedicine
Okada R,Furusawa A,Vermeer DW,Inagaki F,Wakiyama H,Kato T,Nagaya T,Choyke PL,Spanos WC,Allen CT,Kobayashi H
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Sat May 01 00:00:00 UTC 2021
Milk-derived extracellular vesicles alleviate ulcerative colitis by regulating the gut immunity and reshaping the gut microbiota.
Theranostics
Tong L,Hao H,Zhang Z,Lv Y,Liang X,Liu Q,Liu T,Gong P,Zhang L,Cao F,Pastorin G,Lee CN,Chen X,Wang JW,Yi H
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Jan 28 00:00:00 UTC 2022
Hybrid Gene Origination Creates Human-Virus Chimeric Proteins during Infection.
Cell
Ho JSY,Angel M,Ma Y,Sloan E,Wang G,Martinez-Romero C,Alenquer M,Roudko V,Chung L,Zheng S,Chang M,Fstkchyan Y,Clohisey S,Dinan AM,Gibbs J,Gifford R,Shen R,Gu Q,Irigoyen N,Campisi L,Huang C,Zhao N,Jones JD,van Knippenberg I,Zhu Z,Moshkina N,Meyer L,Noel J,Peralta Z,Rezelj V,Kaake R,Rosenberg B,Wang B,Wei J,Paessler S,Wise HM,Johnson J,Vannini A,Amorim MJ,Baillie JK,Miraldi ER,Benner C,Brierley I,Digard P,Łuksza M,Firth AE,Krogan N,Greenbaum BD,MacLeod MK,van Bakel H,Garcìa-Sastre A,Yewdell JW,Hutchinson E,Marazzi I
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Thu Jun 25 00:00:00 UTC 2020
Protein arginine methyltransferase 5 promotes cholesterol biosynthesis-mediated Th17 responses and autoimmunity.
The Journal of clinical investigation
Webb LM,Sengupta S,Edell C,Piedra-Quintero ZL,Amici SA,Miranda JN,Bevins M,Kennemer A,Laliotis G,Tsichlis PN,Guerau-de-Arellano M
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2020
Glufosinate constrains synchronous and metachronous metastasis by promoting anti-tumor macrophages.
EMBO molecular medicine
Menga A,Serra M,Todisco S,Riera-Domingo C,Ammarah U,Ehling M,Palmieri EM,Di Noia MA,Gissi R,Favia M,Pierri CL,Porporato PE,Castegna A,Mazzone M
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Oct 07 00:00:00 UTC 2020
Notch2 complements Notch1 to mediate inductive signaling that initiates early T cell development.
The Journal of cell biology
Romero-Wolf M,Shin B,Zhou W,Koizumi M,Rothenberg EV,Hosokawa H
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon Oct 05 00:00:00 UTC 2020
Enhanced alleviation of aGVHD by TGF-β1-modified mesenchymal stem cells in mice through shifting MΦ into M2 phenotype and promoting the differentiation of Treg cells.
Journal of cellular and molecular medicine
Wu R,Liu C,Deng X,Chen L,Hao S,Ma L
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
Immunoprophylactic and immunotherapeutic control of hormone receptor-positive breast cancer.
Nature communications
Buqué A,Bloy N,Perez-Lanzón M,Iribarren K,Humeau J,Pol JG,Levesque S,Mondragon L,Yamazaki T,Sato A,Aranda F,Durand S,Boissonnas A,Fucikova J,Senovilla L,Enot D,Hensler M,Kremer M,Stoll G,Hu Y,Massa C,Formenti SC,Seliger B,Elemento O,Spisek R,André F,Zitvogel L,Delaloge S,Kroemer G,Galluzzi L
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Thu Jul 30 00:00:00 UTC 2020
Mitochondrial Pyruvate Carrier 1 Promotes Peripheral T Cell Homeostasis through Metabolic Regulation of Thymic Development.
Cell reports
Ramstead AG,Wallace JA,Lee SH,Bauer KM,Tang WW,Ekiz HA,Lane TE,Cluntun AA,Bettini ML,Round JL,Rutter J,O'Connell RM
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Western Blot
Tue Mar 03 00:00:00 UTC 2020
PTPN2 Deficiency Enhances Programmed T Cell Expansion and Survival Capacity of Activated T Cells.
Cell reports
Flosbach M,Oberle SG,Scherer S,Zecha J,von Hoesslin M,Wiede F,Chennupati V,Cullen JG,List M,Pauling JK,Baumbach J,Kuster B,Tiganis T,Zehn D
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Jul 28 00:00:00 UTC 2020
Artificial Mini Dendritic Cells Boost T Cell-Based Immunotherapy for Ovarian Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Cheng S,Xu C,Jin Y,Li Y,Zhong C,Ma J,Yang J,Zhang N,Li Y,Wang C,Yang Z,Wang Y
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2020
Distinct actions of the fermented beverage kefir on host behaviour, immunity and microbiome gut-brain modules in the mouse.
Microbiome
van de Wouw M,Walsh AM,Crispie F,van Leuven L,Lyte JM,Boehme M,Clarke G,Dinan TG,Cotter PD,Cryan JF
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon May 18 00:00:00 UTC 2020
B7-CD28 co-stimulation modulates central tolerance via thymic clonal deletion and Treg generation through distinct mechanisms.
Nature communications
Watanabe M,Lu Y,Breen M,Hodes RJ
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2020
Characterization of immune cell subtypes in three commonly used mouse strains reveals gender and strain-specific variations.
Laboratory investigation; a journal of technical methods and pathology
Hensel JA,Khattar V,Ashton R,Ponnazhagan S
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2019
Antitumor and immunomodulatory effects of a novel multitarget inhibitor, CS2164, in mouse hepatocellular carcinoma models.
Anti-cancer drugs
Zhou Y,Fu C,Kong Y,Pan D,Wang Y,Huang S,Li Z,Ning Z,Lu X,Shan S,Xin L
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
Sex Differences in Mouse Popliteal Lymph Nodes.
Scientific reports
Dill-Garlow R,Chen KE,Walker AM
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Jan 30 00:00:00 UTC 2019
SCGN deficiency results in colitis susceptibility.
eLife
Sifuentes-Dominguez LF,Li H,Llano E,Liu Z,Singla A,Patel AS,Kathania M,Khoury A,Norris N,Rios JJ,Starokadomskyy P,Park JY,Gopal P,Liu Q,Tan S,Chan L,Ross T,Harrison S,Venuprasad K,Baker LA,Jia D,Burstein E
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Wed Oct 30 00:00:00 UTC 2019
Anti-IL-6 eluting immunomodulatory biomaterials prolong skin allograft survival.
Scientific reports
Uehara M,Li X,Sheikhi A,Zandi N,Walker B,Saleh B,Banouni N,Jiang L,Ordikhani F,Dai L,Yonar M,Vohra I,Kasinath V,Orgill DP,Khademhosseini A,Annabi N,Abdi R
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Thu Apr 25 00:00:00 UTC 2019
Hippo Kinases Mst1 and Mst2 Sense and Amplify IL-2R-STAT5 Signaling in Regulatory T Cells to Establish Stable Regulatory Activity.
Immunity
Shi H,Liu C,Tan H,Li Y,Nguyen TM,Dhungana Y,Guy C,Vogel P,Neale G,Rankin S,Feng Y,Peng J,Tao W,Chi H
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Tue Nov 20 00:00:00 UTC 2018
MTOR signaling is essential for the development of thymic epithelial cells and the induction of central immune tolerance.
Autophagy
Liang Z,Zhang L,Su H,Luan R,Na N,Sun L,Zhao Y,Zhang X,Zhang Q,Li J,Zhang L,Zhao Y
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2019
Targeted calcium influx boosts cytotoxic T lymphocyte function in the tumour microenvironment.
Nature communications
Kim KD,Bae S,Capece T,Nedelkovska H,de Rubio RG,Smrcka AV,Jun CD,Jung W,Park B,Kim TI,Kim M
Published figure using CD25 monoclonal antibody (Product # 51-0251-82) in Flow Cytometry
Mon May 15 00:00:00 UTC 2017