Product References
A novel approach for transforming breast cancer stem cells into endothelial cells.
Experimental and therapeutic medicine
Mao QQ,Ji XC,Zhang JN,Teng WF,Zhou SC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2024
Tumor-derived mesenchymal progenitor cell-related genes in the regulation of breast cancer proliferation.
Gland surgery
Chen Y,Zhu L,Wang Y,Hu J,Zhang H,Zhu J,Gong W,Liu X,Xiao F,Li X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Wed Mar 27 00:00:00 UTC 2024
Epigenetic landscape reveals MECOM as an endothelial lineage regulator.
Nature communications
Lv J,Meng S,Gu Q,Zheng R,Gao X,Kim JD,Chen M,Xia B,Zuo Y,Zhu S,Zhao D,Li Y,Wang G,Wang X,Meng Q,Cao Q,Cooke JP,Fang L,Chen K,Zhang L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Tue Apr 25 00:00:00 UTC 2023
MiR-451a and let-7i-5p loaded extracellular vesicles attenuate heme-induced inflammation in hiPSC-derived endothelial cells.
Frontiers in immunology
Thomas JJ,Harp KO,Bashi A,Hood JL,Botchway F,Wilson MD,Thompson WE,Stiles JK,Driss A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Wed Jan 11 00:00:00 UTC 2023
Angiogenic stem cell delivery platform to augment post-infarction neovasculature and reverse ventricular remodeling.
Scientific reports
Shin HS,Thakore A,Tada Y,Pedroza AJ,Ikeda G,Chen IY,Chan D,Jaatinen KJ,Yajima S,Pfrender EM,Kawamura M,Yang PC,Wu JC,Appel EA,Fischbein MP,Woo Y,Shudo Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Thu Oct 20 00:00:00 UTC 2022
Autophagy modulates endothelial junctions to restrain neutrophil diapedesis during inflammation.
Immunity
Reglero-Real N,Pérez-Gutiérrez L,Yoshimura A,Rolas L,Garrido-Mesa J,Barkaway A,Pickworth C,Saleeb RS,Gonzalez-Nuñez M,Austin-Williams SN,Cooper D,Vázquez-Martínez L,Fu T,De Rossi G,Golding M,Voisin MB,Boulanger CM,Kubota Y,Muller WA,Tooze SA,Nightingale TD,Collinson L,Perretti M,Aksoy E,Nourshargh S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunocytochemistry
Tue Sep 14 00:00:00 UTC 2021
Simultaneously targeting cancer-associated fibroblasts and angiogenic vessel as a treatment for TNBC.
The Journal of experimental medicine
Sharma M,Turaga RC,Yuan Y,Satyanarayana G,Mishra F,Bian Z,Liu W,Sun L,Yang J,Liu ZR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunocytochemistry
Mon Apr 05 00:00:00 UTC 2021
Simultaneously targeting cancer-associated fibroblasts and angiogenic vessel as a treatment for TNBC.
The Journal of experimental medicine
Sharma M,Turaga RC,Yuan Y,Satyanarayana G,Mishra F,Bian Z,Liu W,Sun L,Yang J,Liu ZR
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunocytochemistry
Mon Apr 05 00:00:00 UTC 2021
Modeling of endothelial cell dysfunction using human induced pluripotent stem cells derived from patients with end-stage renal disease.
Kidney research and clinical practice
Kim KW,Shin YJ,Kim BM,Cui S,Ko EJ,Lim SW,Yang CW,Chung BH
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunocytochemistry
Wed Dec 01 00:00:00 UTC 2021
Expression and role of interleukin-1β and associated biomarkers in deep vein thrombosis.
Experimental and therapeutic medicine
Pai RZ,Fang Q,Tian G,Zhu B,Ge X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunohistochemistry
Mon Feb 01 00:00:00 UTC 2021
Placental endovascular extravillous trophoblasts (enEVTs) educate maternal T-cell differentiation along the maternal-placental circulation.
Cell proliferation
Ma Y,Yang Q,Fan M,Zhang L,Gu Y,Jia W,Li Z,Wang F,Li YX,Wang J,Li R,Shao X,Wang YL
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Wed Jul 01 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Tue Nov 17 00:00:00 UTC 2020
Both Human Hematoma Punctured from Pelvic Fractures and Serum Increase Muscle Resident Stem Cells Response to BMP9: A Multivariate Statistical Approach.
Journal of clinical medicine
Alinejad Y,Lauzon MA,Grenier G,Balg F,Faucheux N
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Sun Apr 19 00:00:00 UTC 2020
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Tue Dec 08 00:00:00 UTC 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Tue Dec 22 00:00:00 UTC 2020
Matrine inhibits the development and progression of ovarian cancer by repressing cancer associated phosphorylation signaling pathways.
Cell death & disease
Zhang X,Hou G,Liu A,Xu H,Guan Y,Wu Y,Deng J,Cao X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Immunohistochemistry
Thu Oct 10 00:00:00 UTC 2019
Expansion of functional personalized cells with specific transgene combinations.
Nature communications
Lipps C,Klein F,Wahlicht T,Seiffert V,Butueva M,Zauers J,Truschel T,Luckner M,Köster M,MacLeod R,Pezoldt J,Hühn J,Yuan Q,Müller PP,Kempf H,Zweigerdt R,Dittrich-Breiholz O,Pufe T,Beckmann R,Drescher W,Riancho J,Sañudo C,Korff T,Opalka B,Rebmann V,Göthert JR,Alves PM,Ott M,Schucht R,Hauser H,Wirth D,May T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Thu Mar 08 00:00:00 UTC 2018
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 62-0319-42) in Flow Cytometry
Mon May 01 00:00:00 UTC 2017