    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
              • Aspire Member Program
              • Connect: Lab, Data, Apps
            Loading ...
            • Primary Antibodies ›
            • TBC1D9 Antibodies
            Bethyl Laboratories
            TBC1D9 Polyclonal Antibody
            View all (5) TBC1D9 antibodies
            Questions & Answers
            Cite TBC1D9 Polyclonal Antibody
            • Antibody Testing Data (1)
            Application
            TBC1D9 Antibody in Immunoprecipitation (IP)
            Group 53 Created with Sketch.
            FIGURE: 1 / 1
            TBC1D9 Antibody (A301-027A-T) in IP
            Detection of human TBC1D9 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from HEK293T cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-TBC1D9 antibody (Product # A301-027A) (lot 1) used for IP at 6 µg per reaction. TBC1D9 was also immunoprecipitated by rabbit anti-TBC1D9 antibody A301-028A. For blotting immunoprecipitated TBC1D9, A301-028A was used at 1 µg/mL. Detection: Chemiluminescence with an exposure time of 3 minutes.
            View Product
            TBC1D9 Antibody in Immunoprecipitation (IP)
            Product Details
            A301-027A-T
            Applications
            Tested Dilution
            Publications
            Western Blot (WB)
            Assay-dependent
            -
            Immunoprecipitation (IP)
            2-10 µg/mg lysate
            -
            Product Specifications
            Species Reactivity
            Human
            Host/Isotype
            Rabbit / IgG
            Class
            Polyclonal
            Type
            Antibody
            Immunogen
            Region between residue 250 and 300 of Human TBC1 domain family, member 9.
            View immunogen
            Conjugate
            Unconjugated
            Form
            Liquid
            Concentration
            1 mg/mL
            Purification
            Antigen affinity chromatography
            Storage buffer
            tris citrate/phosphate, pH 7-8
            Contains
            0.09% sodium azide
            Storage conditions
            4°C
            Shipping conditions
            Ambient (domestic); Wet ice (international)
            Product Specific Information
            The recommended shelf life for this product is 1 year from date of receipt.
            Target Information
            TBC1D9 may act as a GTPase-activating protein for Rab family protein(s).
            Bioinformatics
            Protein Aliases: TBC1 domain family member 9; TBC1 domain family member 9A; TBC1 domain family, member 9 (with GRAM domain)
            View more View less
            Gene Aliases: GRAMD9; KIAA0882; MDR1; TBC1D9; TBC1D9A
            View more View less
            UniProt ID: (Human) Q6ZT07
            View more View less
            Entrez Gene ID: (Human) 23158
            View more View less
            Function(s)
            GTPase activator activity calcium ion binding protein binding Rab GTPase binding GTPase-activating protein
            Process(es)
            intracellular protein transport regulation of vesicle fusion activation of GTPase activity
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            • Price & Freight Policy
            • Cookie Preferences
              Your choices regarding cookies on this site
              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States
            Your items have has been added!
            Host server : magellan-search-green-584499df9d-8sqwf:80/100.66.74.130:80.
            git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.33.0-2025.07.30-1.0