Search Thermo Fisher Scientific
Immunogen sequence: MSACGRKAL TLLSSVFAVC GLGLLGIAVS TDYWLYLEEG VIVPQNQSTE IKMSLHSGLW RVCFLAGEER GRCFTIEYVM PMNTQLTSES TVNVLKMIRS ATPFPLVSLF FMFIGFILNN IGHIRPHRTI LAFVSGIFFI LSGLSLVVGL VLYISSINDE MLNRTKDAET YFNYKYGWSF AFAAISFLLT ESAGVMSVYL FMKRYTAEDM YRPHPGFYRP RLSNCSDYSG QFLHPDAWVR GRSPSDISSE ASLQMNSNYP ALLKCPDYDQ MSSSPC (1-275 aa encoded by B C101607)
L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: calcium channel, voltage-dependent, gamma subunit 5; Neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; Transmembrane AMPAR regulatory protein gamma-5; Voltage-dependent calcium channel gamma-5 subunit
基因别名: CACNG5
UniProt ID: (Human) Q9UF02
Entrez Gene ID: (Human) 27091