Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Sequence of this protein is as follows: MGTTASTAQQ TVSAGTPFEG LQGSGTMDSR HSVSIHSFQS TSLHNSKAKS IIPNKVAPVV ITYNCKEEFQ IHDELLKAHY TLGRLSDNTP EHYLVQGRYF LVRDVTEKMD VLGTVGSCGA PNFRQVQGGL TVFGMGQPSL LGFRRVLQKL QKDGHRECVI FCVREEPVLF LRADEDFVSY TPRDKQNLHE NLQGLGPGVR VESLELAIRK EIHDFAQLSE NTYHVYHNTE DLWGEPHAVA IHGEDDLHVT EEVYKRPLFL QPTYRYHRLP LPEQGSPLEA QLDAFVSVLR ETPSLLQLRD AHGPPPALVF SCQMGVGRTN LGMVLGTLIL LHRSGTTSQP EAAPTQAKPL PMEQFQVIQS FLRMVPQGRR MVEEVDRAIT ACAELHDLKE VVLENQKKLE GIRPESPAQG SGSRHSVWQR ALWSLERYFY LILFNYYLHE QYPLAFALSF SRWLCAHPEL YRLPVTLSSA GPVAPRDLIA RGSLREDDLV SPDALSTVRE MDVANFRRVP RMPIYGTAQP SAKALGSILA YLTDAKRRLR KVVWVSLREE AVLECDGHTY SLRWPGPPVA PDQLETLEAQ LKAHLSEPPP GKEGPLTYRF QTCLTMQEVF SQHRRACPGL TYHRIPMPDF CAPREEDFDQ LLEALRAALS KDPGTGFVFS CLSGQGRTTT AMVVAVLAFW HIQGFPEVGE EELVSVPDAK FTKGEFQVVM KVVQLLPDGH RVKKEVDAAL DTVSETMTPM HYHLREIIIC TYRQAKAAKE AQEMRRLQLR SLQYLERYVC LILFNAYLHL EKADSWQRPF STWMQEVASK AGIYEILNEL GFPELESGED QPFSRLRYRW QEQSCSLEPS APEDLL
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Paladin; palladin
Gene Aliases: KIAA1274; PALD; PALD1
UniProt ID: (Human) Q9ULE6
Entrez Gene ID: (Human) 27143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support