Search Thermo Fisher Scientific
FIGURE: 1 / 1
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human ASAM/ACAM.
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.
Protein Aliases: Adipocyte adhesion molecule; adipocyte-specific adhesion molecule; CAR-like membrane protein; Coxsackie- and adenovirus receptor-like membrane protein; CXADR-like membrane protein; FLJ22415
Gene Aliases: ACAM; ASAM; CLMP; CSBM; CSBS; UNQ318/PRO363
UniProt ID: (Human) Q9H6B4
Entrez Gene ID: (Human) 79827