Product References
The Antitumor Effect of Caffeic Acid Phenethyl Ester by Downregulating Mucosa-Associated Lymphoid Tissue 1 via AR/p53/NF-κB Signaling in Prostate Carcinoma Cells.
Cancers
Chang KS,Tsui KH,Hsu SY,Sung HC,Lin YH,Hou CP,Yang PS,Chen CL,Feng TH,Juang HH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Thu Jan 06 00:00:00 EST 2022
WNT1 Inducible Signaling Pathway Protein 1 Is a Stroma-Specific Secreting Protein Inducing a Fibroblast Contraction and Carcinoma Cell Growth in the Human Prostate.
International journal of molecular sciences
Chang KS,Chen ST,Sung HC,Hsu SY,Lin WY,Hou CP,Lin YH,Feng TH,Tsui KH,Juang HH
42-6200 was used in Western Blotting to elucidate that WISP1 is a stroma-specific secreting protein, enhancing the cell migration and contraction of prostate fibroblasts, as well as the proliferation, invasion, and tumor growth of prostate carcinoma cells.
Wed Sep 28 00:00:00 EDT 2022
Mucosa-Associated Lymphoid Tissue 1 Is an Oncogene Inducing Cell Proliferation, Invasion, and Tumor Growth via the Upregulation of NF-κB Activity in Human Prostate Carcinoma Cells.
Biomedicines
Tsui KH,Chang KS,Sung HC,Hsu SY,Lin YH,Hou CP,Yang PS,Chen CL,Feng TH,Juang HH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Wed Mar 03 00:00:00 EST 2021
Prostate-derived ets factor represses tumorigenesis and modulates epithelial-to-mesenchymal transition in bladder carcinoma cells.
Cancer letters
Tsui KH,Lin YH,Chung LC,Chuang ST,Feng TH,Chiang KC,Chang PL,Yeh CJ,Juang HH
42-6200 was used in western blot to analyze repression of tumorigenesis and modulation of epithelial-to-mesenchymal transition in bladder carcinoma cells by prostate-derived ets factor
Sat May 28 00:00:00 EDT 2016
The Iron Chelator, Dp44mT, Effectively Inhibits Human Oral Squamous Cell Carcinoma Cell Growth in Vitro and in Vivo.
International journal of molecular sciences
Lee JC,Chiang KC,Feng TH,Chen YJ,Chuang ST,Tsui KH,Chung LC,Juang HH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Wed Aug 31 00:00:00 EDT 2016
N-myc downstream-regulated gene 1 (NDRG1) mediates pomegranate juice protection from apoptosis in hypoxic BeWo cells but not in primary human trophoblasts.
Placenta
Chen B,Zaveri PG,Longtine MS,Nelson DM
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Immunofluorescence
Sat Aug 01 00:00:00 EDT 2015
N-myc downstream-regulated gene 1 (NDRG1) mediates pomegranate juice protection from apoptosis in hypoxic BeWo cells but not in primary human trophoblasts.
Placenta
Chen B,Zaveri PG,Longtine MS,Nelson DM
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Immunofluorescence
Sat Aug 01 00:00:00 EDT 2015
Growth differentiation factor-15: a p53- and demethylation-upregulating gene represses cell proliferation, invasion, and tumorigenesis in bladder carcinoma cells.
Scientific reports
Tsui KH,Hsu SY,Chung LC,Lin YH,Feng TH,Lee TY,Chang PL,Juang HH
42-6200 was used in western blot to analyze growth differentiation factor-15 as a p53- and demehtylation-upregulating gene that represses invasion, cell proliferation, and tumorigenesis in bladder carcinoma cells
Fri Aug 07 00:00:00 EDT 2015
N-myc downstream-regulated gene 1 downregulates cell proliferation, invasiveness, and tumorigenesis in human oral squamous cell carcinoma.
Cancer letters
Lee JC,Chung LC,Chen YJ,Feng TH,Juang HH
42-6200 was used in immunocytochemistry and western blot to determine the role and function of NDRG1 in oral squamous cell carcinoma
Sun Dec 28 00:00:00 EST 2014
Metallothionein 3: an androgen-upregulated gene enhances cell invasion and tumorigenesis of prostate carcinoma cells.
The Prostate
Juang HH,Chung LC,Sung HC,Feng TH,Lee YH,Chang PL,Tsui KH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Tue Oct 01 00:00:00 EDT 2013
Mechanisms by which interleukin-6 attenuates cell invasion and tumorigenesis in human bladder carcinoma cells.
BioMed research international
Tsui KH,Wang SW,Chung LC,Feng TH,Lee TY,Chang PL,Juang HH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Wed Dec 04 00:00:00 EST 2013
Upregulation of prostate-derived Ets factor by luteolin causes inhibition of cell proliferation and cell invasion in prostate carcinoma cells.
International journal of cancer
Tsui KH,Chung LC,Feng TH,Chang PL,Juang HH
Published figure using NDRG1 polyclonal antibody (Product # 42-6200) in Western Blot
Fri Jun 15 00:00:00 EDT 2012
L-Mimosine blocks cell proliferation via upregulation of B-cell translocation gene 2 and N-myc downstream regulated gene 1 in prostate carcinoma cells.
American journal of physiology. Cell physiology
Chung LC,Tsui KH,Feng TH,Lee SL,Chang PL,Juang HH
42-6200 was used in western blot to investigate the molecular mechanisms of L-mimosine on cell cycle modulation in prostate carcinoma cells.
Wed Feb 15 00:00:00 EST 2012
Glycoprotein transmembrane nmb: an androgen-downregulated gene attenuates cell invasion and tumorigenesis in prostate carcinoma cells.
The Prostate
Tsui KH,Chang YL,Feng TH,Chang PL,Juang HH
42-6200 was used in western blot to investigate the function and regulation of glycoprotein transmembrane nmb in the human prostate.
Sat Sep 15 00:00:00 EDT 2012