Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclophilin-like protein PPIL6; peptidyl-prolyl cis-trans isomerase-like variant a; peptidylprolyl isomerase (cyclophilin)-like 6; PPIase; Probable inactive peptidyl-prolyl cis-trans isomerase-like 6; radial spoke 12 homolog; Rotamase PPIL6
Gene Aliases: bA425D10.6; dJ919F19.1; PPIase; PPIL6; RSPH12
UniProt ID: (Human) Q8IXY8
Entrez Gene ID: (Human) 285755
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support