Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Discover the exceptional performance of the Attune Flow Cytometer family—Attune NxT, Attune CytPix, and Attune Xenith—engineered to set a new standard in flow cytometry. At the heart of each Attune Flow Cytometer lies our revolutionary acoustic focusing technology, which utilizes sound waves to meticulously align cells within the flow cell. This innovative approach surpasses traditional hydrodynamic methods, resulting in faster processing speeds, exceptional data quality, and increased sample throughput.
The Attune NxT cytometer is the reliable workhorse instrument your lab can rely on for consistent and dependable performance. The Attune CytPix model features a high-speed brightfield camera and AI-driven image analysis software, with recent upgrades including machine learning tools and an intuitive user interface, enabling enhanced data precision, accuracy, and faster insights into sample biology. The Attune Xenith cytometer is a high-parameter spectral analyzer, providing both spectral unmixing and conventional flow capabilities for comprehensive and versatile cell analysis.
Leverage the power of advanced technology and innovation to accelerate your discoveries. Trust in the Attune Flow Cytometer family to deliver the reliability, speed, and accuracy that set your work apart.
Try our configurator tool to design an Attune NxT or CytPix system to fit your research needs.
For Research Use Only. Not for use in diagnostic procedures.