Multiplexing the TUNEL signal with GFP and RFP
HeLa cells were transduced with CellLight® Mitochondria-GFP, BacMam 2.0 (green) and CellLight® Golgi-RFP, BacMam 2.0 (red). After overnight incubation the cells were subjected to DNase treatment and analyzed for DNA breaks using one of the indicated TUNEL assay kits. When compared to the available TUNEL assays offered by either Roche or Promega, only the Click-iT® Plus Alexa Fluor® 647 assay can simultaneously multiplex both fluorescent proteins.
HeLa cells were transduced with CellLight® Mitochondria-GFP, BacMam 2.0 (green) and CellLight® Golgi-RFP, BacMam 2.0 (red).   After overnight incubation the cells were subjected to DNase treatment and analyzed for DNA breaks using one of the indicated TUNEL assay kits. When compared to the available TUNEL assays offered by either Roche or Promega, only the Click-iT® Plus Alexa Fluor® 647 assay can simultaneously multiplex both fluorescent proteins.
Related Products
Catalog # Name Size Price (USD) Qty
C10617 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
849.65
Online Exclusive
878.00
Save 28.35 (3%)
C10618 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
836.65
Online Exclusive
864.00
Save 27.35 (3%)
C10619 Click-iT™ Plus TUNEL Assay Kits for In Situ Apoptosis Detection, 1 kit Each
839.65
Online Exclusive
868.00
Save 28.35 (3%)
Related Images