  • Contact
Accelerating ScienceAnalyzing Metals / Manufacturing Quality Assurance / What Is Stainless Steel? Part I
What Is Stainless Steel? Part I
Written by Chris Burnett | Published: 01.19.2023
Part I: An Introduction to the World’s Most Popular Metal
portrait of man view from washing machine inside
“Stainless” steel is actually a generic term referring to a variety of steel types. Like all other kinds of steel, stainless steel is made primarily from iron and carbon in a two-step process. What makes stainless steel different is the addition of chromium (Cr) and other alloying elements such as nickel (Ni) to create a corrosion-resistant product.
Steel corrodes because iron, the metal used to make steel, occurs in nature in combination with other elements. When iron ore is artificially manipulated into a pure form to make steel, it becomes unstable and will readily recombine with oxygen.
When chromium is added to steel, it forms chromium oxide, which acts as a protective surface to prevent air and moisture from causing rust, as happens with ordinary steel. Chromium is added in quantities ranging from 10.5 to 30%, depending on the application or environment in which the steel is to be used. There are more than 100 different grades of stainless steel but they can be grouped into five major types:
Austenitic is the most widely used type of stainless steel. It has excellent corrosion and heat resistance with good mechanical properties over a wide range of temperatures. Austenitic steel is used in housewares, industrial piping and vessels, construction, and architectural facades.
Ferritic stainless steel has similar properties to mild steel (the most common steel), but better corrosion, heat, and cracking resistance. Ferritic steel is commonly used in washing machines, boilers and indoor architecture.
Martensitic stainless steel is very hard and strong, though it is not as resistant to corrosion as austenitic or ferritic grades. It contains approximately 13% chromium and is used to make knives and turbine blades.
Duplex stainless steel is a composite of austenitic and ferritic steels, making it both strong and flexible. Duplex steels are used in the paper, pulp, shipbuilding, and petrochemical industries. Newer duplex grades are being developed for a broader range of applications.
Martensitic or semi-austenitic steels can also be classified as precipitation hardening stainless steels. These steels are made to be extremely strong with the addition of elements such as aluminum, copper and niobium.
Corrosion resistance is the main advantage of stainless steel, but it certainly isn’t the only one. Stainless steel is also:
  • High and low temperature resistant
  • Easily fabricated
  • Strong and durable
  • Easy cleaned and maintained
  • Long lasting, with a low lifecycle cost
  • Aesthetically attractive
  • Environmentally friendly and recyclable.
In addition to chromium, stainless steels are made with alloys of silicon, nickel, carbon, nitrogen, and manganese. Nitrogen, for example, improves tensile properties like ductility. Nickel is added to austenitic steel to improve flexibility. These alloys are added in varying amounts and combinations to meet specific end-use applications, which is why it’s very important for stainless steel manufacturers to verify that the correct percentages of each alloy are being used. There are two technologies that provide the elemental analysis needed to produce high quality stainless steel: X-Ray Fluorescence (XRF) and Optical Emission Spectroscopy (OES).
Handheld XRF and LIBS are highly valuable technologies in the scrap metal market. Stainless steel is 100% recyclable and therefore a huge amount of stainless steel must be evaluated at the scrap yard. Handheld XRF analyzers bring immense value to scrap metal recyclers because it is a highly accurate, nondestructive testing technique that can analyze a metal sample in seconds with little to no need for sample preparation.  With XRF, stainless steel scrap can be quickly analyzed and sorted according to grade and type. Laser Induced Breakdown Spectroscopy (LIBS) is better for carbon analysis for metals and alloys.
OES is a robust, reliable, and widely-used technology for the analysis of metals and alloys in the lab. Compared with traditional combustion analyzers, OES provides faster elemental analysis with high precision and accuracy in iron and steel, aluminum, copper, magnesium, precious metals and other specialty metals/alloys. OES has demonstrated its capability to provide more efficient control of steel production by providing accurate sample analysis during the manufacturing process.
Additional Resources:
  • Download our free eBook: A Practical Guide to Improving Steel Manufacturing Processes and Production Methods
  • Read more about how the multitude of stainless steel grades are grouped into five primary classifications in our blog article:  What is Stainless Steel? Part II
  • Visit our center for Improving Steel Manufacturing Processes and Production
visit our improving steel processes center
Editor’s Note:  This article was previously published February 18, 2014, but has been refreshed and links updated.
Share this article
381
SHARES
FacebookLinkedinTwitterMail
Detail of a corporate jet fuselage, rivets, and engine
Could a Metal Screw or Fastener Affect Airplane Safety?
With the media focused lately on the airplanes, helicopters,... by Peter Anzalone / 04.22.2025
Read More
The pipe and valve in oil fields
Residual Elements in Piping can Cause Big Problems in Refineries; How Elemental Analysis can Help
Ensuring material integrity in refining operations is critic... by Mark Lang / 12.10.2024
Read More
A tag with "Happy Thanksgiving" printed on it is tied to a small pumpkin that isits next to a dinner plate set for a Thanksgiving dinner.
Happy Thanksgiving to our Analyzing Metals Readers
We want to say thank you this week to all those workers who ... by Marlene Gasdia-Cochrane / 11.26.2024
Read More
Say Goodbye to Sulphur Surprises in Medical Steel
Sulphur is typically considered an impurity in medical-grade... by Mark Lang / 11.19.2024
Read More
scrap metal recycling ebook
eBook: A Practical Guide to Improving Metal and Alloy Sorting for Scrap Metal Recyclers
Download Guide
Download free ebook for steel manufacturing
eBook: A Practical Guide to Improving Steel Manufacturing
Download Guide
Comments
  1. Steel and Site says
    January 13, 2020 at 12:09 pm
    Good information about stainless steel. It is an informative blog post for professionals in the steel industry.
    Reply
  2. Shaylee Packer says
    February 11, 2020 at 1:40 pm
    I didn’t realize that stainless steel was created as a steel option that wouldn’t rust. We are looking to put some steel beams in the backyard. It seems like, as you mentioned, it would be best for us to get stainless steel. This way, it will last longer and stay looking nice.
    Reply
  3. Al Mutqan says
    June 1, 2020 at 10:27 am
    Great introduction about stainless steel metal fabrication.
    Stainless steel is one of the material that is being used by all the industry for their equipment manufacturing.
    Thanks for sharing the information.
    Reply
  4. Dorothy Marie Wood says
    July 22, 2020 at 10:10 am
    I must find out what alloy is possibly used in medical operations … stainless steel made with what alloys? Is it an industrial secret involving a patient allergic to Chromium and Nickel and possibly other metals specifically used in medical insertions into the body, securing bone breakage with screws. It should not be propietary under the circumstances a life is in jeopardy for a relatively simple break. .
    Reply
  5. sandeep singh says
    July 24, 2020 at 6:42 am
    nice article. thanks for sharing this knowledge with us. this is very useful for me.
    Reply
  6. Rosie Beckett says
    September 30, 2020 at 6:34 pm
    My little sister is competing in her first go-cart race in a few months, and one of the requirements to enter is to build your own car. She is thinking about using stainless steel fasteners to help keep the parts together, and I didn’t realize that steel has so many benefits! I really appreciate you explaining that stainless steel is strong and durable, so it won’t bend to the weight of large loads.
    Reply
    • Jeff says
      September 14, 2021 at 6:25 pm
      Great Point Rosie. We are huge fans of Stainless Steel, especially since we are located in the Florida Panhandle where humidity and saltwater is an issue for rust and corrosion.
      Reply
  7. manpreet singh says
    November 3, 2020 at 8:32 am
    This is such a good article. I always read these types of articles and gaining knowledge from them. Thanks for sharing valuable knowledge with us.
    Reply
  8. Marine Parts Auckland says
    December 17, 2020 at 8:12 am
    It is a nice article about three reasons about what is stainless steel part. I agree with all your points that you have stated here, love this blog. You have done a great research for I feel, thanks for sharing.
    Reply
  9. Brad Nailers says
    December 24, 2020 at 8:15 am
    I really appreciate you for publishing this blog here about what is stainless steel part-i; it’s really a helpful and very useful for us. This is really appreciated that you have presented this data over here, I love all the information shared. Great article!
    Reply
  10. *Eliza says
    February 9, 2021 at 12:48 pm
    *Many people which are not from industrial backgrounds believe that stainless steel can’t get rusty and that stainless steel is completely rust-proof. In fact, although stainless steel is a metal that contains chromium and other elements offering a certain amount of resistance to corrosion the base material is virtually as active as ordinary carbon steel.
    Reply
  11. aluminium fabrication Auckland says
    April 7, 2021 at 8:53 am
    Your article is quite helpful! I have so many questions, and you have answered many. Thank you! Such a nice and superb article, we have been looking for this information about what is stainless steel part i. Indeed a great post about it!!
    Reply
  12. Neha sharma says
    June 21, 2021 at 9:56 am
    Great post! Thanks for sharing wonderful information.
    Reply
  13. Fabrication says
    August 21, 2021 at 2:18 pm
    Great article. thanks for sharing this knowledge with us. this is very helpful for me.
    Reply
  14. Tuan Nguyen Huu says
    September 10, 2021 at 3:59 am
    Great! Thanks for sharing.
    Reply
  15. Jeff says
    September 15, 2021 at 8:42 pm
    Great info Chris Burnett.
    Reply
  16. Jeff says
    September 16, 2021 at 1:32 pm
    Hope your sister has a great experience there Rose. Stainless Steel is a great option for many uses, but for heavy duty parts (keeping the wheels on), you still might want to look at traditional steel bolts as they can have drastically different strengths.
    Reply
  17. Jeff says
    September 17, 2021 at 12:40 pm
    Thanks for sharing. Lots of info and things to think about.
    Reply
  18. Jeff says
    September 17, 2021 at 6:51 pm
    Well written article by Chris Burnett. Thank you. I keep forgetting about the benefits of Stainless Steel recycling and semi environmentally friendly. Great reminder.
    Reply
  19. Jeff Beeblebrox says
    September 30, 2021 at 6:06 pm
    Great article. Thank you for taking your time to write this.
    Reply
  20. Fabrication In Jaipur says
    November 11, 2021 at 9:16 am
    What a blog? It really useful post. I am searching for this information for a few days and I found this information few days and I found this information on your blog. Your blog is very informative. Thanks for sharing this information with us.
    Reply
  21. michin mu says
    February 9, 2022 at 12:14 pm
    This is an authoritative source for all the information on stainless steel, thank you for letting me know more
    Reply
  22. visit us says
    March 3, 2022 at 8:00 pm
    I keep forgetting about the benefits of Stainless Steel recycling and semi environmentally friendly. Great reminder.
    Reply
  23. Enggprosolutions says
    March 14, 2022 at 8:54 am
    Useful information. Keep sharing!!!
    Reply
  24. http://www.tacomaconcretecontractors.com/ says
    March 31, 2022 at 7:16 pm
    Fantastic! This article is good and informative content that gives valuable information
    Reply
  25. website says
    April 4, 2022 at 8:09 pm
    You have remarked very interesting points. Many Thanks for sharing.
    Reply
  26. Website says
    April 14, 2022 at 7:54 pm
    Excellent work for creating this kind of content.
    Reply
  27. our site says
    May 1, 2022 at 2:43 pm
    Your blog is very informative. Thanks for sharing this information with us.
    Reply
  28. Digital Aditya says
    June 11, 2022 at 7:06 am
    Beautifully Explained. Such nice blog thanks for it
    Reply
  29. Ausbilderschein onlinekurs says
    November 28, 2022 at 1:51 pm
    very nice article. thanks.
    Reply
  30. Rohit says
    October 28, 2024 at 6:45 am
    This is a great introduction to stainless steel! I really appreciate the clear explanation of what stainless steel is and the importance of chromium in the alloy. Could you explain more about how the percentage of chromium affects the corrosion resistance in different types of stainless steel?
    Reply
    • Marlene Gasdia-Cochrane says
      December 18, 2024 at 9:16 pm
      Hi. Thanks for asking. We have written several articles about corrosion. Here’s one:
      XRF Remains an Important Tool to Combat Steel Pipe Corrosion
      Additionally, visit our Steel Center web pages and you will find a variety of educational materials about steel production: https://www.thermofisher.com/us/en/home/industrial/manufacturing-processing/steel-manufacturing/resources.html
      Reply
  31. Santhana Lakshmi Metals says
    January 6, 2025 at 8:06 am
    Comment:
    This article provides an excellent introduction to the various types of stainless steel and their applications across different industries. The detailed explanation of how chromium and other alloying elements give stainless steel its unique corrosion resistance and strength is particularly insightful. It’s also fascinating to learn about the role of technologies like XRF and OES in ensuring the quality of stainless steel during production. As stainless steel continues to be crucial in many sectors, manufacturers like Sanlak are contributing greatly to ensuring that the right alloy compositions are met to produce high-quality stainless steel for diverse uses. With its recyclability and numerous advantages, stainless steel truly stands out as one of the most versatile materials available today!
    Reply
  32. Santhana Lakshmi Metal says
    January 27, 2025 at 5:43 am
    This article explains stainless steel really well. It’s interesting how adding chromium makes it resistant to rust, and the different types of stainless steel are used in everything from kitchenware to construction. The recycling part is cool too, since it’s 100% recyclable and can be easily sorted with technology like XRF.
    Reply
Trackbacks
  1. Is Plastic Pipe The Answer to Combating Corrosive Steel in Hydrocarbon Processing? - Analyzing Metals says:
    May 3, 2016 at 7:48 pm
    […] Carbon dioxide may be trapped in oil reservoirs or present in crude oil as a by-product, but usually it’s injected into crude oil wells for enhanced oil recovery. CO2 causes sweet corrosion, which typically results in pitting or material loss. Sweet corrosion can be eliminated by adding inhibitors to the crude oil or by using stainless steel pipe. […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Celebration and Thanks to our Analyzing Metals Readers
Answering 5 Questions from Metal Fabrication Manufacturers
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook