Search
Cell Factory Systems, multilayered cell culture flasks, are a proven solution for scaling adherent cell cultures from laboratory to production volumes. Designed to provide high surface area within a compact footprint, these multilayer systems enable efficient expansion while maintaining consistency with traditional flask-based workflows.
Thermo Scientific Nunc Cell Factory systems use the same surface treatments as Nunc flasks and are compatible with closed fluid management solutions, helping reduce contamination risk and support CGMP manufacturing.
Compare an example workflow replacing 850 cm² roller bottles with Nunc Cell Factory 40 systems to estimate potential net savings and impacts on labor, consumable costs, facility footprint, and CO₂ emissions.
Nunc EasyFill |
Nunc Standard |
Nunc High Density |
|
|
|
Primary benefit |
Fast, simple filling and handling |
Flexible multilayer system for adherent cell scale-up |
Higher surface area in the same footprint |
Shorter lead time |
Customizable closed system designed for your specific process |
System type |
Open system with wide fill opening |
Open or configurable |
Open multilayer system |
Fully closed system with tubing and filters |
Fully closed system using industry-standard components (tubing, connectors, vent filters) |
Surface area efficiency |
Standard growth surface |
Standard growth surface |
Up to 30% more surface area compared to other Nunc Cell Factories due to reduced vertical space between layers |
Standard growth surface |
Standard growth surface |
Available formats |
1, 2, 4, 10, 40 layers |
1, 2, 4, 10, 40 layers |
3, 13, 52 layers |
2, 10 layers |
2, 4, 10, and 40 layer configurations |
Best suited for |
Drug discovery and R&D |
Process development |
Manufacturing |
Process development |
Manufacturing
|
A multilayered flask like a Nunc Cell Factory System may be considered when expanding adherent cells. Multiple layers improve space utilization in incubators and reduce handling since fewer vessels are used. They can support efficient seed train expansion, vaccine production workflows, viral substrate generation, and intermediate adherent culture steps.
Selection typically begins with the total surface area required to meet yield targets, manufacturing stage, and filling preferences. For example, EasyFill formats are a good choice for research applications while standard closed systems are useful for GMP or contamination sensitive applications. For maximizing incubator space, high-density options have reduced vertical space between layers allowing more layers within the same space.
Scaling from flasks to multilayer systems can maintain similar growth profiles when surface treatment and material consistency are aligned. However, differences in gas exchange, media distribution, and handling across layers may influence performance. Start by selecting systems with the same surface treatment across multiple cell culture systems like the Nunclon Delta surface treatment. Other factors like ease of cell harvest may need to be optimized.
Harvesting typically involves introducing dissociation reagents through designated ports or connectors, followed by controlled incubation to promote detachment. Closed or semi-closed tubing options can support fluid transfer. Gentle agitation using shaker platforms or automated manipulators may assist in uniform reagent distribution and cell release, depending on workflow and scalability requirements.
Contamination control strategies often include closed-system handling, sterile connectors, and filtered venting options to limit exposure during liquid transfers. Minimizing open interventions, performing open interventions within a sterile hood, and standardizing handling procedures can help improve batch-to-batch consistency.
Roller bottles and microcarriers are alternative options for adherent cell expansion. Microcarriers, beads that support cell growth, can be used with systems like the Thermo Scientific HyPerforma Single-Use Bioreactors for scalable production. As processes move toward GMP, closed systems become essential, and Thermo Fisher Scientific can design fluid transfer solutions to fit your needs.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.