Family of Gibco High-Intensity Perfusion CHO Medium
What's new—Gibco Efficient-Pro Medium & Feeds help enhance CHO cell protein quality and productivity
Gibco Efficient-Pro Medium and Gibco Efficient-Pro Feeds 1 and 2 help deliver superior product performance, improved product titer, better protein quality and lot-to-lot consistency, which provides more control and predictability. Each medium is chemically defined, completely scalable and available as Gibco Advanced Granulation Technology (AGT) formats for sustained cell viability. The AGT dry media format enables a simple and scalable reconstitution process. These formats help maximize process efficiency with simplified preparation. Customers with larger-scale operations will benefit from longer bioreactor runs, reduced risk in reconstitution and flexibility to support these CHO cell lines.
Gibco Efficient-Pro Medium in liquid and Advanced Granulation Technology (AGT) formats
Gibco Efficient-Pro™ Medium
  • Tested in, and pairs best with feeds for, CHO-S, CHO-K1, and CHO-DG44
  • Together cell-specific Efficient-Pro Feeds, helps increase titer and cell viability
Gibco Efficient-Pro Feeds 1/ Gibco Efficient-Pro Feeds 1in liquid and Advanced Granulation Technology (AGT) formats
Gibco Efficient-Pro™ Feed 1
  • Tested in, and pairs best with feeds for CHO-K1
  • Together with Efficient-Pro Medium, helps increase titer and cell viability
Gibco Efficient-Pro Feeds 1/ Gibco Efficient-Pro Feeds 1in liquid and Advanced Granulation Technology (AGT) formats
Gibco Efficient-Pro™ Feed 2
  • Tested in, and pairs best with feeds CHO-S and DG44
  • Together with Efficient-Pro Medium, helps increase titer and cell viability
CHO cells cultured in Efficient-Pro media demonstrated improved protein production—High titers established across CHO variants and vessel size show increased end-product yield, viable cell density, reduced batch numbers, and lower cost with CHO-K1 and CHO-S cells.
Glycan and charge variant profiles—Improved protein quality established across all CHO variants creating increased end-product yield, reduced purification steps, and improved efficacy in CHO-K1 and CHO-S cells.
High specific productivity of IgG across nearly all CHO variants and vessel size demonstrates opportunity for improved yield, enhanced protein production, and extended harvest stage in CHO-K1, CHO-S, and DG-44 cells.
CHO-K1 cell growth
Efficient-Pro Medium and Efficient-Pro Feed 1 maintained high viability and lower cell density indicating a higher specific productivity compared to the competitor.
CHO-S cell productivity
Compared to the alternative medium and feeds, Efficient-Pro Medium and Feed 2 with CHO-S cells realized with a 144% relative average titer by day 14 and up to 180% relative average specific productivity (qP).
DG-44 cell productivity
Compared to the alternative medium and feeds, Efficient-Pro Medium and Feed 2 with DG44 cells showed a 312% relative average titer by day 14 with up to a 160% relative average specific productivity (qP).
CHO cells cultured in Efficient-Pro media demonstrated improved protein production—High titers established across CHO variants and vessel size show increased end-product yield, viable cell density, reduced batch numbers, and lower cost with CHO-K1 and CHO-S cells.
Glycan and charge variant profiles—Improved protein quality established across all CHO variants creating increased end-product yield, reduced purification steps, and improved efficacy in CHO-K1 and CHO-S cells.
High specific productivity of IgG across nearly all CHO variants and vessel size demonstrates opportunity for improved yield, enhanced protein production, and extended harvest stage in CHO-K1, CHO-S, and DG-44 cells.
CHO-K1 cell growth
Efficient-Pro Medium and Efficient-Pro Feed 1 maintained high viability and lower cell density indicating a higher specific productivity compared to the competitor.
CHO-S cell productivity
Compared to the alternative medium and feeds, Efficient-Pro Medium and Feed 2 with CHO-S cells realized with a 144% relative average titer by day 14 and up to 180% relative average specific productivity (qP).
DG-44 cell productivity
Compared to the alternative medium and feeds, Efficient-Pro Medium and Feed 2 with DG44 cells showed a 312% relative average titer by day 14 with up to a 160% relative average specific productivity (qP).
Learn more about AGT dry media formats and their advantages
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Learn more about available media formats designed to support optimal CHO cell culture
      High-Intensity Perfusion CHO Medium
      Gibco High-Intensity Perfusion (HIP) CHO Medium can be used at multiple concentrations to support your process from seed train to production.
      Just as it can be used at multiple concentrations, it can be used in multiple perfusion processes: N-1 and seed train, intensified fed-batch, concentrated fed batch, and continuous perfusion. Numerous CHO cell lines (CHO-S, CHO-DG44, CHO-GS, CHO-K1) have been tested in these perfusion processes with HIP CHO Medium.
      In addition, the medium supports high cell densities at low medium exchange rates, which improves process efficiency, as low as 1 vessel volume/day (VVD).
      Learn more about HIP CHO ›
      Key features of HIP CHO Medium
      Features
      Benefits
      Advanced granulated technology (AGT) format
      Easy to use
      Works with numerous CHO clones
      CHO-S, CHO-GS, CHO-K1, CHO-DG44
      Works in a variety of processes
      Intensified fed-batch, concentrated fed batch, N-1, and continuous perfusion
      Supports high cell densities at low medium exchange rates
      Improved process efficiency
      Supports multiple reconstituted concentrations
      Product can be flexibly reconstituted to meet process needs
      Gibco ExpiCHO SPMGibco ExpiCHO SMP
      Gibco ExpiCHO SPM eliminates guesswork needed to find compatible media and feeds for scale-up. Simply transition your ExpiCHO-S cells after stable transfection for the same high titer and virtually none of the optimization.
      • Seamless transition
      • Confidence in performance
      • Risk reduction
      Ideal with:
      Cells
      ExpiCHO-S
      Feeds
      EfficientFeed C+
      Protein Quality
      GlycanTune Total Feeds
      Request a sample ›
      Learn more about the ExpiCHO family ›
      ExpiCHO family includes
      • ExpiCHO expression system for transient expression
      • ExpiCHO-S cells
      • ExpiCHO SPM
      All products are designed to work seamlessly with one another easing the transition to stable expression helping you to commercialization faster.
      Provides you with:
      • ONE solution—no media optimization required
      • Hassle-free scale-up—stable clone development protocol available
      Learn more about the ExpiCHO family ›
      expicho-family
      The ExpiCHO SPM formulation is optimized for the ExpiCHO-S cells and outperforms other CHO medium on the market.
      Figure 1 illustrates a comparative analysis. Stable clones were developed using the ExpiCHO stable clone development protocol and then transitioned to test media for an Ambr15 run. The highest output was achieved by ExpiCHO SPM, with the liquid and AGT (dry) formats performing comparably. The data represents 16 replicates for each format of ExpiCHO SPM and 3 replicates each for ActiPro and Advanced CHO respectively.
      Efficient-Pro Medium and Feeds System
      The Gibco Efficient-Pro Medium and Feed System is designed to drive productivity in your batch or fed-batch CHO process. This system includes state-of-the-art basal medium and carefully optimized single-part feeds designed specifically for our cell line.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          High-Intensity Perfusion CHO Medium
          Gibco High-Intensity Perfusion (HIP) CHO Medium can be used at multiple concentrations to support your process from seed train to production.
          Just as it can be used at multiple concentrations, it can be used in multiple perfusion processes: N-1 and seed train, intensified fed-batch, concentrated fed batch, and continuous perfusion. Numerous CHO cell lines (CHO-S, CHO-DG44, CHO-GS, CHO-K1) have been tested in these perfusion processes with HIP CHO Medium.
          In addition, the medium supports high cell densities at low medium exchange rates, which improves process efficiency, as low as 1 vessel volume/day (VVD).
          Learn more about HIP CHO ›
          Key features of HIP CHO Medium
          Features
          Benefits
          Advanced granulated technology (AGT) format
          Easy to use
          Works with numerous CHO clones
          CHO-S, CHO-GS, CHO-K1, CHO-DG44
          Works in a variety of processes
          Intensified fed-batch, concentrated fed batch, N-1, and continuous perfusion
          Supports high cell densities at low medium exchange rates
          Improved process efficiency
          Supports multiple reconstituted concentrations
          Product can be flexibly reconstituted to meet process needs
          Gibco ExpiCHO SPMGibco ExpiCHO SMP
          Gibco ExpiCHO SPM eliminates guesswork needed to find compatible media and feeds for scale-up. Simply transition your ExpiCHO-S cells after stable transfection for the same high titer and virtually none of the optimization.
          • Seamless transition
          • Confidence in performance
          • Risk reduction
          Ideal with:
          Cells
          ExpiCHO-S
          Feeds
          EfficientFeed C+
          Protein Quality
          GlycanTune Total Feeds
          Request a sample ›
          Learn more about the ExpiCHO family ›
          ExpiCHO family includes
          • ExpiCHO expression system for transient expression
          • ExpiCHO-S cells
          • ExpiCHO SPM
          All products are designed to work seamlessly with one another easing the transition to stable expression helping you to commercialization faster.
          Provides you with:
          • ONE solution—no media optimization required
          • Hassle-free scale-up—stable clone development protocol available
          Learn more about the ExpiCHO family ›
          expicho-family
          The ExpiCHO SPM formulation is optimized for the ExpiCHO-S cells and outperforms other CHO medium on the market.
          Figure 1 illustrates a comparative analysis. Stable clones were developed using the ExpiCHO stable clone development protocol and then transitioned to test media for an Ambr15 run. The highest output was achieved by ExpiCHO SPM, with the liquid and AGT (dry) formats performing comparably. The data represents 16 replicates for each format of ExpiCHO SPM and 3 replicates each for ActiPro and Advanced CHO respectively.
          Efficient-Pro Medium and Feeds System
          The Gibco Efficient-Pro Medium and Feed System is designed to drive productivity in your batch or fed-batch CHO process. This system includes state-of-the-art basal medium and carefully optimized single-part feeds designed specifically for our cell line.
          Video Player is loading.
          Current Time 0:00
          Duration 0:00
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 0:00
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
              Gibco media and feed selections by cell line
              This table provides information on the most likely candidate based on a specific cell line. As all clones are unique, other media may in fact work more ideally and it is suggested to try a variety of media to test.
              For more information on considerations when selecting the optimal match, go to our next section.
              CHO cell typeGibco media optionMatched feed optionDoes medium contain growth factor? (Y/N)Is adaption necessary? (Y/N)
              CHO-S
              Efficient-Pro Medium
              Efficient-Pro Feed 2
              N
              Y
              CHO Medium
              Difco TC Yeastolate UF
              Y
              Y
              Dynamis Medium
              EfficientFeed C+
              N
              Y
              ExpiCHO-S
              ExpiCHO SPM w/gf add back
              EfficientFeed C+
              Y
              Y
              ExpiCHO SPM
              EfficientFeed C+
              N
              N
              DG44/DXB11
              Efficient-Pro Medium
              Efficient-Pro Feed 2
              N
              Y
              CHO Medium
              OneFeed Supplement
              Y
              Y
              Dynamis or CD OptiCHO Medium
              EfficientFeed C+ or EfficientFeed A+
              N
              N
              CHO-K1
              Efficient-Pro Medium
              Efficient-Pro Feed 1
              N
              Y
              Dynamis Medium
              EfficientFeed C+
              N
              Y
              GS CHO
              CD-1000 Medium
              Resurge CD5 Supplement
              Y
              Y
              Dynamis or CD CHO
              EfficientFeed C+ or EfficientFeed B+
              N
              Y
              Other CHO
              CHO Medium
              Difco TC Yeastolate UF
              Y
              Y
              Dynamis or CD CHO Medium
              EfficientFeed B+ or EfficientFeed C+
              N
              Y
              *Drug Master File (DMF) information may be available and can be found at Drug Master Files (DMFs) and Regulatory Support Files (RSFs).
              Find your optimal match with Gibco cell culture media
              There are several pathways to find the right media or media and feed solution for CHO cell cultures. These pathways include selecting a standard or off-the-shelf media, our CHO media test panel, or a media development or optimization service. In order to find the right path, we need to remove the biggest obstacle; luck.
              Often time, your goals on what you want to accomplish will narrow down the best selections for you.
              Questions to consider:
              • What are you hoping to accomplish?
              • Do you know your timeline?
              • What is most critical?
              Comparison of the different options available
              Standard catalog media
              Media test panel
              Development or optimization service
              Definition
              Standard catalog products include our off-the-shelf CHO media, feed and supplement options.
              Developed media panels with diverse formulations offering improved specificity, without the need for a full media development service.
              Service solutions are provided by our Gibco BioProduction Services team, offering cell line development, media development, media optimization, process optimization, consultations, prototyping, format conversions and more.
              Time
              Fast
              Relative speed
              Dependent on scope; >3 months
              Budget
              Economical
              Mostly cost-effective
              More investment needed
              Titer
              Average to meets requirements
              Meets requirements
              Meets or surpasses requirements
              Benefits
              Quick turn-around
              Standard media are available off-the-shelf.
              Easy scale-up
              Liquid and dry formats available that provide performance consistency and may reduce workflow steps to simplify your process.
              Risk mitigation and assurance of supply
              May be produced at either of our harmonized manufacturing locations.
              Available data
              Frequently, used in application notes and further testing.
              Capacity
              Secondary sourcing options; easy scale-up
              More specificity
              Options that are more specific to your clone and molecule.
              Relative speed
              Speed is important but not as much as matching product quality.
              Regulatory support
              For the panels, provides regularity support needed to succeed in getting approvals.
              Resources and/or expertise
              Need to outsource or consultation on your process
              Secondary sourcing
              Increase capacity of your own media and mitigate risk
              Scale-up solutions
              Transitioning from a liquid to a dry format during scale-up or performance support during scale-up or tech transfer
              Improved performance
              Optimal solution that delivers on your performance criteria
              Platform media
              Options that work better than current platform for multiple molecules
              Regulatory support
              We offer royalty-free CHO cell lines and documentation for regulatory filings.
              Frequently, cell line specific component dependencies, like insulin and other growth factors, are missed when evaluating media. These are components that are needed for your cell line to thrive (growth, productivity) and if they are not present in the tested media, you may miss out on the optimal solution.
              Matching media to your cell line should include an evaluation of whether these dependencies are in the media of choice or not (Tables 1 and 2) to determine whether you need to add them in or not.
              Even with the cell line dependency component, in most cases, adaptation is required to see optimal results.
              Additional components may also need to be considered for an optimal outcome (Table 3).
              Questions:
              • Do you know if your cell line has any dependencies?
              • Are you planning on adapting the cells to the media?
              • Are other considerations necessary for the best result?
              Standard catalog media
              Media test panel
              Development or optimization service
              Dependencies
              Table 1 lists media containing growth factors
              Table 2 lists media without growth factors
              Our media test panel does not contain typical cell line dependencies by default and can be added in as requested.
              Our service offerings will evaluate these factors and other factors as specified.
              Considerations
              Additional components may be necessary for optimal performance (Table 3).
              Table 1. Products that contain a growth factor, by cell line.
              Cell type
              Media option
              Matched feed option
              CHO-S
              CHO Medium
              Difco TC Yeastolate UF
              ExpiCHO-S
              ExpiCHO SPM* w/gf add back
              EfficientFeed C+
              DG44/DXB11
              NutriCHO CD Medium* EX, LX, MAX
              OneFeed Supplement
              GS CHO
              CD-1000 Medium
              Resurge CD5 Supplement
              Other CHO
              CHO Medium
              Difco TC Yeastolate UF
              ALL
              Development or optimization Services and Media Test Panel
              Based on specifications
              * No cell adaptation required
              Table 2. Products that do not contain a growth factor, by cell line.
              Cell type
              Media option
              Matched feed option
              CHO-S
              Dynamis Medium
              EfficientFeed C+ Supplement
              ExpiCHO-S
              ExpiCHO SPM*
              EfficientFeed C+ Supplement
              DG44/DXB11
              CD OptiCHO Medium*
              EfficientFeed A+ Supplement
              CHO-K1
              Dynamis Medium
              EfficientFeed C+ Supplement
              GS CHO
              Dynamis Medium
              CD CHO Medium
              EfficientFeed C+ Supplement
              EfficientFeed B+ Supplement
              Other CHO
              Dynamis Medium
              EfficientFeed C+ Supplement
              All
              Development and optimization services and Media Test Panel
              Based on specifications
              * No cell adaptation required
              Table 3. Troubleshooting components to bundle with your medium selection.
               Situation
              Recommendation
              Cell Line cloned in medium containing insulin
              Insulin and/or Long R3 IGF
              Cell line is clumping
              Titrate Anti-Clumping Agent starting at 1%
              Cell line is sensitive to ammonia
              GlutaMAX Supplement
              Cell line is not sensitive to ammonia
              Glutamine 
              Performance drops (productivity, apoptosis), especially during scaling up
              Peptones
              Need to adjust the glycosylation profile
              GlycanTune A+, B+ or C+ Total Feeds or GlycanMod Pak
              Not all CHO clones are alike; each clone has a different metabolic baseline. When determining a new medium for your clone, you may want to consider a variety of available medium on the market to test. All options are scalable from small- to large-scale in a variety of formats.
              Standard catalog media
              Media test panel
              Development or optimization service
              Options
              Order samples based on the below groupings for a diverse panel:
              No growth factors in medium
              • Dynamis
              • CD CHO
              • CD OptiCHO
              • ExpiCHO SPM
              Containing growth factors in medium
              • CHO Medium
              • CD-1000 Medium
              • NutriCHO CD
              Order ›
              Panel consists of five diverse chemically defined media which can be mixed to produce 21 test conditions
              • Completion of several design of experiments with panel formulations
              • Formulations can be easily supplemented upon request
              • Flexibility to perform experiments at your facility or ours
              Learn more ›
              In partnership with you, this service will provide solutions that are specific to your requirements.
              Learn more ›
              CHO media—ordering information
              Catalog # Name Size Price (USD) Qty
              A5221601 Efficient-Pro™ AGT™ Feed 2, 1 L Each
              387.00
              A5221602 Efficient-Pro™ AGT™ Feed 2, 10 L Each
              3,255.00
              The flexibility of Gibco CHO media and feeds give you options to pair with most industry utilized cell lines, and are available in both catalog and customized options in multiple formats to suit your application needs:  
              • Faster growth
              • Highest performing titers
              • Sustained viable cell density
              • Chemically defined, serum-free, protein-free, and animal origin-free (AOF)
              • Flexible formats: liquid and AGT
              • Options to customize for every medium
              Related products and services
              Bioprocessing resources
              Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
              For Research Use or Further Manufacturing. Not for diagnostic use or direct administration in to humans or animals.