Search Thermo Fisher Scientific
The Thermo Scientific ClipTip tips are a excellent tool designed to enhance precision and ease in laboratory pipetting tasks. With its unique ClipTip technology, the tips enable secure attachments, reducing the risk of tip ejection and sample loss. This innovative system combines accuracy, reliability, and user comfort to deliver exceptional results in a wide range of application.
ClipTip system | Friction sealing systems |
---|---|
Rely on mechanism designed clips on the pipette nozzle that securely hold the tip in place. The technology eliminates the risk of tip ejection, helping reduce the potential for sample loss and contamination. | Rely on a tight fit between the pipette tip and pipette nozzle. The tip is pushed onto the nozzle until it forms a friction seal. Some variation in the force required to attach the tip, which can lead to inconsistent results. |
![]() |
![]() |
ClipTip security |
Fit for Purpose | Color-coded tip racks |
![]() |
![]() |
|
ClipTip tips remain sealed until you decide to eject them. Enabling consistent and reproducible pipetting. |
Our commitment to supporting your scientific achievements begins with ensuring that you can rely on products to be of high available quality and to meet or exceed your expectations for performance according to the requirements of your applications. |
ClipTip tips racks are colored coded allowing the end user select the right tip volume for the application. |
Tip Type |
Features | Recommended Use |
Filtered Tip |
|
|
Non-filtered tip
|
|
|
Packaging tips |
Features and benefits |
|
|
|
|
|
|
Color |
96-format volume range |
384-format volume range |
|
10 µl/20 µl |
12.5 µl |
|
50 µl |
30 µl |
|
100 µl/200 µl |
125 µl |
|
300 µl |
- |
850 µl/1000 µl |
- |
|
|
1250 µl |
- |
ClipTip technology is the excellent solution in which the pipette and tip form a pipetting system that helps increase the confidence in reproducibility, reduces forces required to attach and eject tips, and secures excellent accuracy and precision.
Pipette |
Compatible ClipTip tip |
Features and benefits |
F1 – ClipTip manual pipette
|
20µl, 50µl, 200µl, 300µl, 300µl Ext, 1000µl
|
Interlocking ClipTip technology provides a secure seal on every channel Light tip attachment and ejection forces Locking volume adjustment prevents volume drift Light-weight handle with adjustable finger rest for left- or right-handed use
|
E1- ClipTip electronic pipette
|
96-Format: 12.5µl* and 12.5µl Ext , 200µl, 300µl and 300µl Ext, 1000µl, 1250µl
384-format: 12.5µl and 12.5µl Ext, 30µl, 125µl
|
Versatile solution for liquid handling, begin pipetting immediately with preset function or create a customized, step-based program with Matrix function Interlocking ClipTip technology provides a secure seal on every channel, eliminating variability Equalizer’s adjustable spacing allows rapid transfer between multiple liquid vessel formats (example: tubes to plates) Electronic tip ejection and index finger operation enhances ergonomics and efficiency, reducing hand strain Connect Bluetooth enabled models to My Pipette Creator application for centralized, efficient programming and data storage and sharing |
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.