1x1 image pixel for data collection
SEM EDS with ChemiSEM Technology
SEM with energy dispersive X-ray spectroscopy for materials characterization.
What is energy dispersive X-ray spectroscopy?
Energy dispersive X-ray spectroscopy (EDS, or EDX) is a powerful analytical technique for characterizing chemical composition on the macro-, micro- and nanoscale. EDS derives compositional information from X-rays emitted when the electron beam scans over a sample, with each element identified from its characteristic X-ray line profile.  EDS is used to analyze almost the entire periodic table, providing information on the distribution of the elements present.  Information provided by EDS supports in-depth material characterization for a wide range of industrial and academic applications, spanning process/quality control to failure analysis and fundamental research. 
What is energy dispersive X-ray spectroscopy SEM and ChemiSEM image of ceramic in glass showing the localization of the zirconium. The ChemiSEM image highlights the presence of aluminum in the area of interest, otherwise not visible with the SEM only.
How ChemiSEM Technology enables EDS analysis
Thermo Scientific ChemiSEM Technology offers a leap forward in usability, convenience, and speed of analysis. It integrates advanced elemental analysis with the electron image in real time. Within one software interface, the operator can:
  • View the sample image from any electron detector.
  • See areas of different chemical composition on the sample surface.
  • Viewpoint and area compositional analyses with a single click.
  • Chart compositional changes across the sample.
  • Create maps of elemental or phase distribution across the sample.
ChemiSEM Technology simplifies EDS analysis by fully integrating SEM and EDS functions into a single, cohesive user interface. Based on live quantification and building on decades of expertise in EDS analysis, the technology provides elemental information quickly and easily, helping to ensure reliable results. And because ChemiSEM Technology is always on, it can dramatically shorten time to results, highlight features that would have previously gone unnoticed, and provide more complete information.
How ChemiSEM Technology enables EDS analysis ChemiSEM makes all the needed information available in one time, allowing users to speed up their work with more complete data.
Benefits of SEM EDS analysis with ChemiSEM Technology
  • Always on—Eliminate the need to move from SEM imaging to EDS analysis for each sample. ChemiSEM Technology is always on and continuously collects the EDS signal during SEM imaging, collecting a full EDS spectrum at every point in the image.
  • Faster data acquisition—Access elemental information much faster than conventional techniques with novel data segmentation approaches based on machine learning, ultrafast signal processing, and the tightest possible SEM EDS integration.
  • Reliable results—Get accurate qualification and quantification over the widest range of operating conditions including beam energy, sample size, and working distance.
  • Complete information—See the big picture with comprehensive micro-scale elemental composition. Immediate color results reveal defects or imperfections you might have otherwise missed.
  • Multi-data viewing—See SEM and elemental information in a single frame for complete characterization of your sample at a glance.
  • Simplified operation—Immediately view compositional results that make elemental information accessible to everyone and easily increase the number of scientists or engineers who can use your facilities.
ChemiSEM Technology is designed and built to provide full control over all SEM and EDS functions within a single user interface. Experiment setup, image processing, and spectral analysis functions are intuitively grouped, making data management straightforward. And the system features a quadrant format and project tree to highlight the most important data.
ChemiPhase analysis: Identify unique phases using advanced statistical analysis
ChemiPhase is a new ChemiSEM Technology feature that allows to run phase analysis on areas of interest by using a big data approach to detect all statistically significant spectra within the datacube. Characterizing the presence of different phases in complex materials is now much more straightforward and intuitive. ChemiPhase is a comprehensive, unbiased statistical engine. Compared to traditional methods that are highly dependent on assumptions about the samples, ChemiPhase avoids erroneous results if unexpected elements are missed due to overlapping peaks or insufficient intensities.
Benefits of ChemiPhase analysis
  • No user bias; fully automated process with no prior identification of elements.
  • Complete and comprehensive analysis; major and minor components identification down to a single pixel with no peak overlaps.
  • Fast and reliable; phase determination with very little X-ray data (as few as 10 counts per pixel) and results provided in less than a minute, even for complex phase maps.
  • Reduced complexity; image and spectrum view pair for each distinct phase. Automatic optimization of all spectra for a more confident analysis.
Meet the scientists using ChemiSEM Technology
ChemiSEM Testimonial - Carlos Angeles Chavez
Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
        1. Now Playing
          Up NextChemiSEM Testimonial - Carlos Angeles Chavez
          Carlos Angeles Chavez from the Mexican Petroleum Institute comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
        2. Now Playing
          Up NextChemiSEM Testimonial - Lifeng Dong
          Lifeng Dong from Hamline University comments on his first impressions of his live demo with the Thermo Scientific™ Prisma E SEM with ChemiSEM technology.
        3. Now Playing
          Up NextChemiSEM Testimonial - Cedric Barroo
          Cedric Barroo from the Universite Libre de Bruxelles comments on his first impressions of his live demo with the Thermo Scientific Prisma E SEM with ChemiSEM technology.
        4. Now Playing
          Up NextChemiSEM Testimonial - Houston Electron Microscopy
          Dennis and Marina Manuel comment on the time saving and productivity benefits that they experienced in their live demonstration of the Thermo Scientific Prisma with ChemiSEM Technology.
      ChemiSEM Technology applications
      Stainless landing gear in Axia ChemiSEM
      The Thermo Scientific Axia ChemiSEM provides high-quality imaging of steel samples to aid in the production of high-value steels.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
            1. Now Playing
              Up NextStainless landing gear in Axia ChemiSEM
              The Thermo Scientific Axia ChemiSEM provides high-quality imaging of steel samples to aid in the production of high-value steels.
            2. Now Playing
              Up NextChlorine-rich contaminant characterization on poorly washed etched steel
              Characterization of a contaminated surface of etched steel conducted with Thermo Scientific ChemiSEM Technology to study the root cause of the contamination. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            3. Now Playing
              Up NextContaminant characterization on Li-ion cathode material
              A thorough assessment of the contaminants within battery materials is critical to assure battery quality and performance using the Thermo Scientific Axia ChemiSEM. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            4. Now Playing
              Up NextCopper- and nickel-coated brass cross section
              Copper and nickel layers are easily identified with Thermo Scientific ChemiSEM Technology, which shows that their material contrast, with BSE detection, is similar. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            5. Now Playing
              Up NextCu-Ni coating characterization on aluminum alloy
              Copper and nickel layers are easily identified with Thermo Scientific ChemiSEM Technology, which shows that their material contrast, with BSE detection, is similar. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            6. Now Playing
              Up NextBend failure mechanism of high-strength steel
              Bend testing is conducted on steel and other materials using Thermo Scientific ChemiSEM Technology to determine ductility and resistance to fracture. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            7. Now Playing
              Up NextFailure analysis of paint defects on a car door cross section
              Thermo Scientific ChemiSEM Technology allows you to investigate the presence of foreign particles in automotive exposed steel panels and to assess their composition. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            8. Now Playing
              Up NextCharacterization of contaminants in brown-fused alumina refractory material for steel production
              Thermo Scientific ChemiSEM Technology characterizes refractory lining materials employed in steelmaking to clarify distribution of phases within final composites. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            9. Now Playing
              Up NextPhase characterization in a zirconia mullite refractory
              Thermo Scientific ChemiSEM Technology characterizes refractory lining materials employed in steelmaking to clarify distribution of phases within final composites. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            10. Now Playing
              Up NextTalc (filler) characterization in a tire cross section
              Thermo Scientific ChemiSEM Technology characterizes a talc particle, one of the most employed fillers in tires. Learn more: https://www.thermofisher.com/nl/en/home/materials-science/chemisem.html
            11. Now Playing
              Up NextThermal barrier coating layer characterization
              The Thermo Scientific Axia ChemiSEM’s large-area imaging makes voids and cracks visible in thermal barrier coatings. Learn more: https://www.thermofisher.com/nl/en/home/electron-microscopy/products/scanning-electron-microscopes/axia-chemisem.html