Bagged enrichment media
Streamline media preparation. Ready. Set. Keep your lab on the go.
Increase productivity by decreasing culture media preparation time with Thermo Scientific™ Bagged Enrichment Media. Designed for media preparation in minutes, this scalable range of solutions eliminates the need for weighing and autoclaving of dehydrated culture media, enabling you to quickly meet testing demands and focus your resources on higher value activities. Work with confidence knowing that your media will be ready to go when you are, and your sample preparation and testing won’t be held up.
Contact us to discuss your lab's needs
FitBag Enrichment Media
Save storage space and preparation time with pre-weighed, ready-to-rehydrate Thermo Scientific™ FitBag™ Enrichment Media. Prepare when convenient: just fill with deionized water, break the seal and mix, for media that’s ready to dispense in under 10 minutes. Reduce waste and increase your confidence of quality with a rehydrated, in use shelf-life of 10 days and certification provided by an ISO/IEC17025 accredited QC laboratory.
QuickBag Enrichment Media
Reduce the need for extra handling and the risk of contamination during enrichment media preparation with Thermo Scientific™ QuickBag™ Media, which are supplied with pre-weighed media and pre-filled water compartments. Just break the seal, mix and start dispensing as needed. QuickBag Media has the added convenience of a validated shelf-life of 10 days once rehydrated, along with the QC release being in accordance with ISO11133:2014 and testing performed in an ISO17025 accredited QC laboratory.
Dry-Bags Enrichment Media
Make bulk enrichment media preparation more convenient with Thermo Scientific™ Dry-Bags™ Media, available in 20 liter format. Simply add deionized water to the media in the pre-filled bag and be ready to dispense in less than half an hour.
Featured videos
FitBag Enrichment Media
Learn how ready-to-fill FitBag Enrichment Media can be quickly and easily prepared when you need it. Store as convenient space-saving bags or store pre-filled with water and ready-to-mix.
Video Player is loading.
Current Time 0:00
Duration 2:27
Loaded: 0%
Stream Type LIVE
Remaining Time 2:27
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextFitBag Enrichment Media
        Learn how ready-to-fill FitBag Enrichment Media can be quickly and easily prepared when you need it. Store as convenient space-saving bags or store pre-filled with water and ready-to-mix.
      2. Now Playing
        Up NextQuickBag Enrichment Media
        Pre-weighed media and pre-filled water compartments of QuickBag™ Enrichment Media could speed your media preparation workflow. Just break the seal, mix and start dispensing as needed.
      3. Now Playing
        Up NextEnrich your efficiency
        Enrich your efficiency with simplified enrichment and sample preparation for food testing laboratories.
    Achieve fast, accurate media dispensing
    Work confidently and accurately: rapidly deliver precise quantities of diluents at the touch of a button with the DiluFlux Pro Automated Gravimetric Dilutors.
    Learn more ›
    Get ready for maximum productivity
    Achieve powerful, reliable homogenization in your sample preparation with the Homogenizer Laboratory Blender. The Homogenizer delivers quiet, efficient blending from enrichment media in Dry-Bags or other formats.
    Learn more ›