1x1 image pixel for data collection
Make way for more powerful imaging
The fully automated EVOS M7000 Imaging System was designed to simplify the most demanding slide and cell-based imaging applications including live-cell analysis, image tiling, and Z-stacking. It is equipped with advanced 2D deconvolution capabilities which increase signal-to-noise ratio (SNR) and provide clean, accurate information, allowing you to reveal hidden insights within your samples.
Video Player is loading.
Current Time 0:00
Duration 0:58
Loaded: 0.00%
Stream Type LIVE
Remaining Time 0:58
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    Features
    In addition to the common features of all EVOS microscopes, the EVOS M7000 Imaging System offers:
    Full automation
    Automated routines help streamline workflow and improve experimental reproducibility
    Speed
    Scan a 96-well plate in 3 fluorescence channels in less than 5 minutes
    2D deconvolution
    Capture images with exceptional clarity and detail
    Time-lapse live-cell imaging
    Optional onstage incubator enables precise control over temperature, humidity, and gas levels
    Area view
    Move rapidly and seamlessly between single-field mode and low- and high-magnification scan modes to easily define and capture the area of interest
    Data analysis
    Seamlessly transfer images to optional
    Celleste Image Analysis Software for access to powerful tools for image segmentation, classification, and cell-based assays
    Virtual 360º view
    Check out the EVOS M7000 Imaging System virtually.
    Explore 360º view
    Note: This tool is best viewed in Chrome, Firefox, or Safari browser.
    Learn more
    EVOS_M7000 instrument
    Advanced 2D deconvolution
    Experience the exceptional clarity and detail of 2D deconvolution.
    Two images side-by-side comparing mouse kidney section before (left) and after (right) deconvolution.
    FluoCells™ Prepared Slide #3 (mouse kidney section with Alexa Fluor™ 488 WGA, Alexa Fluor™ 568 Phalloidin, and DAPI), imaged at 4X (Cat. No. AMEP4922) using EVOS M7000 Imaging System, deconvolved using EVOS Analysis.
    Two images side-by-side comparing U2OS cells before (left) and after (right) deconvolution.
    U2OS cells plated in Nunc™ Glass Bottom Dishes (Cat. No. 150682) and labeled with Hoechst 33342, trihydrochloride trihydrate, 10 mg/mL (Cat. No. H3570, nucleus-blue), Alexa Fluor™ 488 phalloidin (Cat. No. A12379, actin-green), and alpha Tubulin Monoclonal Antibody (236-10501) (Cat. No. A11126) with Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 647 (Cat. No. A21235, tubulin-purple).  Imaged on EVOS M7000 Imaging System with Olympus 100x Objective, semi-apo, 0.95 NA/0.2WD (Cat. No. AMEP4988).
    Two images side-by-side comparing Hela cells before (left) and after (right) deconvolution.
    Hela cells, PFA fixed and stained with DAPI, anti-coilin (GFP), anti-tubulin (Cy5), and Ki-67 (RFP), imaged using EVOS M7000 Imaging System with 20x fluorite objective (Cat. No. AMEP4698), deconvolved using EVOS Analysis software.
    EVOS M7000 HCA Package
    The EVOS M7000 Imaging System, High-Content Analysis Package (AMF7000HCA) combines the fully automated, inverted, multi-channel fluorescence and transmitted light imaging system with Celleste 6 Image Analysis Software, a full-featured image analysis suite. The Celleste 6 Image Analysis software facilitates the transition into low-throughput high-content analyses (HCA) while maintaining the versatility and power of the EVOS M7000 automated microscope. 
    Using plate-based multi-channel analysis (MCA) protocols leveraging machine-learning based algorithms together with its icon-based wizard-driven workflow, Celleste 6 software can help you efficiently segment and classify images for a wide range of assays or bioapplications, including neurite outgrowth, angiogenesis, cell viability, transfection efficiency, and many more. The software also offers a wide range of well-plate data displays including heat maps, image montages, and kinetic graphing options.
    EVOS M7000 & Celleste 6 HCA Packages
    Image shows various bio-applications available with multi-class feature of Celleste
    Featured images and videos
    HeLa cell widefield vs deconvolved
    EVOS M7000, 60x objective, Celleste Image Analysis Software. Stains: NucBlue (nucleus), Phalloidin (F-actin).
    Image displays ability of EVOS M5000 to capture multicolor fluorescent images
    Stitching of rat brain tissue
    EVOS M7000, 4X objective, 16 x 9 stitched images.
    Image displays ability of EVOS M5000 to capture multicolor fluorescent images
    A549 Spheroid
    EVOS M7000, 10X objective. Stains: NucBlue, MitoTracker Orange, CellROX Deep Red.
    Image displays ability of EVOS M5000 to capture multicolor fluorescent images
    Proliferating cells in rat intestine section
    EVOS M7000, color camera.
    Image displays ability of EVOS M5000 to capture multicolor fluorescent images
    Human Cardiac Arterial Smooth Muscle (HCASM) primary cells
    EVOS M5000. 10x objective.
    Image displays ability of EVOS M5000 to capture multicolor fluorescent images
    View more M7000 images and movies in our Image Gallery and Video Gallery.
    Enhance your imaging experiments with optimized product pairings
    Save time with carefully matched product pairings designed to deliver exceptional imaging results.
    Specifications
    For Research Use Only. Not for use in diagnostic procedures.