EVOS S1000 Spatial Imaging System for Multiplexed Tissue Imaging
EVOS S1000 Spatial Imaging System enables 9-plex tissue imaging with confocal-like quality in hours
The EVOS S1000 Spatial Imaging System allows integrated acquisition, processing and high-resolution image generation for spatial localization of proteins in less than one hour. Well-suited for researchers who want to image up to 9 markers in one round, the EVOS S1000 Spatial Imaging System helps provide insights into cellular neighborhoods, interactions and spatial relationships.
Benefits
  • Simple—performs 9-plex tissue imaging in a single-round without bleaching or cycling
  • Fast—produces fully stitched, unmixed, multiplexed images across multiple samples in hours
  • Flexible—compatible with your preferred fluorophores, antibodies, reagents, and with the analysis software of your choice
  • Richer results—capture significantly more data at a fraction of the time of conventional methods
See what sets the EVOS S1000 Spatial Imaging System apart
Spin it. Zoom it. Explore it. Take the interactive 3D tour to explore the innovations behind fast imaging, great flexibility, and exceptional clarity.
More colors. More context. More insights.
Simultaneously capturing and resolving up to eight fluorescence targets plus nuclear stain (9-plex), the EVOS S1000 Spatial Imaging System streamlines the process of tissue imaging and enables powerful image visualization. Compared to conventional imaging (1–4 colors), 9-plex in a single round generates significantly more data for downstream analysis and at a fraction of the cost of cyclic imaging. 
Figure 1. Detecting more targets helps provide more details about the tissue microenvironment and highlights the complexity of biological systems within tissues.  Images of normal colon (left) and adenocarcinoma tissues (right) stained with the 9-plex colon panel on the EVOS S1000 Spatial Imaging System. Multiplex immunofluorescence staining enables information to be collected about the localization and interaction of biomolecules and cells within the tissue microenvironment. 
Figure 2. Multiplex capabilities allow the visualization of more targets in every sample. Unmixed multifield region of an axial Murine kidney FFPE sample labeled with 8 Aluora dyes targeting aquaporins 1, 2, and 4, cytokeratins 8, 18, and 19, MCM2, and smooth muscle actin (SMA) and counterstained with DAPI. Panels 1–4 show the same area of interest and illustrate that increasing the number of labeled targets within a sample produces greater detail. The images were captured using the 20x objective on the Invitrogen EVOS S1000 Spatial Imaging System. 
Table 1. Benefits of multiplex immunofluorescence staining.
Features
With the EVOS S1000 Spatial Imaging System you can:
  • Perform spectral fluorescence, transmitted brightfield, phase-contrast, and color brightfield imaging.
  • Detect intricate multiplexed staining patterns within tissues with a 4.2 megapixel, 16-bit sCMOS camera.
  • Select different types of available objectives (2.5x–40x objectives) that can be accommodated on the 5-position objective slider.
  • Scan, spectrally unmix, stitch and save a 9-plex fluorescence image of a 1 cm2 tissue area at 20x in well under 1 hour.
  • Resolve spectrally overlapping fluorophore signals with automated unmixing that is integrated as part of the image acquisition.
  • Experience navigation, live tissue exploration, spectral extraction, image collection and data retrieval in the simplest possible way, for users with different levels of experience.
  • Generate TIFF or OME-TIFF with single or pyramidal structure available so you can use the data analysis software of your choice.
  • Select and utilize your preferred antibodies and reagents, as the S1000 System is compatible with multiple labeling technologies.
  • Avoid the need for proprietary consumables to reduce your instrument running costs.
Advancing multiplex imaging for spatial proteomics research
Watch and learn about the advancements in multiplex imaging for spatial protein research. Discover how the Invitrogen EVOS S1000 Spatial Imaging System simplifies workflows, supports multiple dyes, and enables fast, high-resolution imaging. Learn about the Invitrogen Aluora Spatial Amplification Reagents and how they provide high sensitivity and bright signals for flexible antibody usage. Understand how fast and simple multiplexing antibodies can be with primary antibodies conjugated to fluorophores and when to use conjugation kits including the Invitrogen ReadyLabel antibody conjugation kits.
Good morning or good evening, everyone, depending on where you are in the world. My name is Adyary Fallarero, and I am a senior manager in product management for imaging technologies at Thermo Fisher Scientific. I'm located in Finland. Hello, everyone. My name is Chris Langsdorf, and I'm a global product manager in protein and cell analysis, also at Thermo Fisher Scientific, and I'm in the United States.
We are both super grateful to have you all attending our talk, and today we will be sharing with you all of the new tools that our company has recently developed to advance multiplex imaging for spatial proteomics research. We hope you'll benefit and gain new knowledge on spatial biology. Feel free to get in contact with us and our teams afterwards if you need any more information or if you have questions.
So let's get started.
We'll jump right in and look at new tools to tackle multiplex imaging challenges in spatial proteomics. Our agenda today includes looking briefly at the revolution in spatial biology, examining some challenges that arise when we start to look at multiplex proteomic imaging, and discussing how we've addressed these challenges using both sample labeling and imaging and analysis techniques.
Spatial biology provides us better insights into our samples primarily because we're able to analyze not just single cells, but also their context in a larger environment. We can use antibody-based detection to tell us what cell types are present and the state of those cells, such as their metabolic or activation state, and what functions they're carrying out. Because our samples are generally fixed, we can get an idea of their identity in both time and space. By studying multiple cell types simultaneously, we can understand the interaction of those cells and how they form larger and more complex neighborhoods or networks.
We can begin to understand both the tissue microenvironment and the architecture of our sample in its native state. Let's look at an example of how this works. We can image an entire tissue section and zoom in on one section of the sample to understand the spatial context. By displaying multiple different colors, we can see the location of different cell types as they exist in their native environment. Then we can use our analysis tools to mask and identify the different cell types that we've labeled with antibody-based detection. We can go a step further with our analysis and ask questions like, "What is the nearest neighbor to each of these cell types?" and begin to understand the network and architecture of our sample in much greater detail.
Several challenges arise when we switch to this type of spatial workflow. Generally, with microscopy, we use primary antibodies followed by fluorescently labeled secondaries. When we move from three or four antibody-based targets up to eight, secondary antibodies are no longer useful. We need to pivot and begin using primary antibody conjugates. As we see here, the primary antibody labeling workflow is very fast. We can make a cocktail of all of these antibodies and label our sample in about an hour. There are some drawbacks, though, such as a generally limited catalog menu available of these conjugates. Unlike with secondary-based detection of three or four targets, where we always choose the three or four brightest dyes available, here we need to choose dyes based on their spectral properties and not necessarily on their brightness.
Fortunately, there's a solution to this challenge provided by the Aluora Spatial Amplification Reagents. Here we use a three-step workflow. First, we label our sample with a primary antibody, followed by a wash and then labeling with a secondary antibody HRP conjugate. After this, we add our Aluora spatial amplification reagents, which are covalently attached to our sample in the location of our antibody binding. After this step, all of the antibodies are stripped off our sample, leaving just a very bright fluorescent labeling in the location of each antigen that was originally present. As we can see on the left, after one round of labeling, we can see the location of one antigen of interest. After repeating, we can see a second, third, fourth, and up to eight plex with great sensitivity and very bright signal. This gives us a lot of flexibility in how we choose and use our antibodies. Because amplification is provided, we can use substantially less antibody, and we're not required to covalently attach a dye to the antibody. The drawback here is it's a longer workflow, so we have to go through multiple labeling cycles.
The Aluora Spatial Amplification Reagents are compatible with primary antibody conjugates. You see two examples here where the primary antibody conjugates are labeled with a few of our brightest dyes available, such as Alexa Fluor 647 and 594, which give us great signal to background for these primary conjugates. With either a less bright dye or a lower expressed antigen, we switch to the Aluora spatial amplification reagent to give us that signal to background that we need. As far as the availability of primary antibody conjugates, there are a few ways besides just acquiring these from a catalog. These are quite available to produce as a DIY method. There are kits available that make it very easy to label your antibody with your dye of choice that's compatible with the system, either by assembling a kit using your own purification system, or we have a technology called ReadyLabel that does two things: it both purifies your antibody and then labels it with one of these eight compatible dyes and gives you a purified conjugate. Or if you ship your antibody to us, our experts will conjugate it and return it back to you. There is a lot of flexibility and convenience available with these options. However, they can be a bit more expensive than just acquiring a conjugated antibody from a catalog.
Thanks so much to Chris for sharing with us the labeling tools that we've been developing to advance spatial proteomics research. Now we're going to switch gears a bit and focus on instrumentation, specifically fluorescent microscopy instrumentation. You all are very aware that fluorescent microscopy can be and has been extensively used to investigate how single cells are organized into a tissue microenvironment, and none of the currently available technologies are really perfect. They all have benefits and drawbacks. What we've tried to do from our perspective is tackle some of the drawbacks of the currently available instrumentations and progress in that manner the type of research that spatial biology scientists can perform. I start maybe with a bit of an overview of what we currently have available and then how we have addressed the current limitations of the existing technologies.
I think you are all very well aware that a great body of research that has been done around spatial proteomics has been done using cyclic labeling and imaging. Imaging that is done in this approach is really pretty conventional. There are between two to four fluorescent channels that are used on those instruments. The benefit of this approach is that because it is performed in a cyclic manner, it allows for a high number of protein targets to be interrogated. The drawback, on the other hand, is that because it's a cyclic process, it tends to be of a lower throughput, and because it is constant illumination of the sample and they go through certain rounds of stripping, that can damage the integrity of the tissue.
The approach that we have taken and that we have tried to address the current limitations is really spectral imaging. Spectral imaging allows for the capture of simultaneous fluorophores within one single round. Instead of only doing the conventional two to four channels, maybe even five fluorescent channels in one single go, the aim of spectral imaging is to capture between seven to ten, even sometimes more, fluorophores in a single round. There is an advantage from doing this: tissue gets better preserved. However, there is also a drawback, and the drawback comes from the fact that when there are so many fluorophores that get acquired in a single imaging round, there is signal bleed-through that needs to be eliminated.
If we try to understand a bit more why this drawback exists in spectral emissions and where it comes from, we can go to the root cause of that problem. The root cause is that the majority of the fluorophores used for spatial imaging have a very broad emission spectrum. If we are imaging ECAD in a 514 channel, the emission spectra of the 514 channel will also appear and show up in both neighboring channels, such as 488. To resolve that signal and remove that bleed-through, we can apply a mathematical process called unmixing. Unmixing is a relatively straightforward process by which we establish the relative contribution that each fluorophore used in that specific protocol has to every pixel of the image. This is done by acquiring spectral signatures from the unstained tissues to account for the typical autofluorescence seen in biological samples, and then from the individually stained fluorophores on the same tissue where the assay is being performed.
Using spectral emission, protein targets can be easily resolved and discriminated even if the fluorescent spectral signals overlap. As you can see in the bottom images shown here, ECAD in the 514 channel can be very clearly discriminated from the CD163 and the PD1 signal that appears in the two neighboring channels. This main drawback associated with spectral technologies is something that we truly wanted to address with our new instrument. Our new instrument is launching now in the fall and is called Invitrogen EVOS S1000 Spatial Imaging System. We have paid a lot of attention to simplifying the unmixing workflow, keeping it very straightforward. It also performs in a way that it is part of the acquisition, done on the fly while the images are being acquired. We have also tried to verify the quality of the unmixing. We provide substantive data as part of a quality report that allows the user to understand the quality of the unmixing that has been performed.
We have gone beyond improving the unmixing workflows by addressing other challenges encountered with currently existing spectral imagers. One of them is the fact that they are often restricted to very specific labeling methods. We have tried to expand the number of dyes that can be utilized and the type of technologies that can be used with EVOS S1000. We have also tried to make it fast so you don't have to spend hours to acquire a 9-plex. You can do it in a single round. EVOS S1000 allows users to acquire up to 8 protein targets and DAPI, making for a 9-plex, while keeping a high resolution of 325 nanometers per pixel at 20x. Last but not least, it's about making spatial proteomics and spatial biology accessible to a broader audience so that many more researchers can utilize spatial biology and study biological processes using these tools. We are not making it simple, but we are trying to make it simpler.
Let's go through the challenges that are presented in currently existing spectral imagers. The first one is that unmixing tends to be limited to very specific dyes that are often proprietary. We have tried to address that issue. When you generate a protocol with EVOS S1000, you will be presented with a repertoire of dyes, roughly around 30 dyes, that includes Alexa Fluor, Alexa Fluor Plus, and special amplification reagents such as Aluora Spatial Amplification Reagents and dyes. You can select the dyes, and the software will show you the overlap that exists on the emission spectra of those dyes that have been selected. We then set up a channel configuration automatically that will acquire the primary channels, shown in blue, as well as the support channels needed to eliminate bleed-through from the proximity of the spectral signals. This is done completely automatically without any user input. The user can also define the product targets on the assay.
Another challenge we have tried to address is the complexity of performing unmixing for anything higher than a 7-plex. Our solution is using the same unmixing workflow regardless of the plex. If you go to a 7-plex or all the way to a 9-plex, the workflow remains the same. First, an unmixing matrix needs to be generated by extracting spectral data from images. Users will need to first acquire an image of their unstained tissue slide and then individual single-color calibrator slides. Once that happens, you can utilize that protocol and start setting up a 9-plex assay. The system will start to acquire images in every single field of view, and at the same time, unmixing will be triggered as part of the image acquisition workflow. You can acquire a 1-square-centimeter 9-plex image that is fully unmixed and stitched in an OMET file format within roughly one hour using a magnification of 20x.
The unmixing workflow eliminates the need for any post-processing. Users should not only subjectively assess the quality of their unmixing. We have addressed this issue by generating an unmixing quality metrics report that contains both raw images versus the unmixed image for qualitative assessment and quantitative parameters that allow users to estimate the quality of the unmixing. We provide clear cutoff values and troubleshooting tips to improve the unmixing process if needed.
We believe that every system and method for microscopy needs to be simple and intuitive, even for users new to the space. We have a proprietary feature called Periscope mode that allows users to turn on any channel and explore their tissue, making it practical and simple for new users to interact with. We aim to provide sophisticated simplicity that allows anyone to quickly gain confidence and easily set up, acquire, and visualize 9-plex images.
The EVOS S1000 can be utilized with multiple labeling strategies. Examples of results obtained using Aluora special amplification reagents show very bright signals that can easily discriminate between targets with low expression levels. The EVOS S1000 can also be used with primary conjugates, allowing for a quick labeling step completed within about two hours at 20x, including unmixing and processing of the data to generate an OME TIFF file. If primary conjugates are not available, another option is to directly conjugate primary antibodies using ReadyLabel kits. This approach is compatible with the EVOS S1000 and provides flexibility in labeling strategies.
The EVOS S1000 Spatial Imaging System is a novel alternative in the spatial imaging space. We aim to provide a choice of achieving high multiplex ability in a single emission step without the complexity of a laborious unmixing workflow. We have made it simple and reliable, providing quality metrics and flexibility in using spatial amplification reagents or validated primary antibody conjugates. We are happy to answer any questions and be a partner in your spatial biology journey. Thank you so much for joining the talk.
Overcome the limitations of traditional immunohistochemistry with the EVOS S1000 Spatial Imaging System and gain more information from your tissues
The EVOS S1000 Spatial Imaging System transforms immunohistochemistry (IHC) by addressing the limitations of single- or double-staining of multiple signals within a single, stained tissue sample. This innovative approach in spatial biology helps EVOS S1000 Spatial Imaging System deliver a comprehensive snapshot of tissue microenvironments and architecture in their native state (Figure 1, above).
Generate highly resolved multiplex immunofluorescence images with automatic unmixing
Benefits of EVOS S1000 Imaging System spectral unmixing technology
The EVOS S1000 Spatial Imaging System software facilitates spectral unmixing automatically whenever is needed. Spectral unmixing is a process used to resolve signals from fluorophores with significant spectral overlap, enabling higher multiplexing than conventional fluorescence microscopy by eliminating the need for spectrally distinct fluorophores and expanding the number of targets that can be resolved in a single imaging acquisition round. (Figure 3).
See spectral unmixing in action with EVOS S1000 Spatial Imaging Software. Slide to toggle Raw and Unmixed.
This unmixing process leverages unique spectral signatures from each fluorophore to determine the abundance of different signal inputs at each pixel, allowing for precise identification and mapping of various fluorophores within an imaged sample. For the algorithm to function effectively, reference spectra are required to extract the spectrum of each fluorophore, which can be obtained using either predicted spectra (default) or measured spectra through careful preparation of single-color controls. Additionally, an unstained sample is necessary to define the tissue's autofluorescence, which is extracted as an independent spectral signature alongside the fluorophores used in the experiment.
Figure 4. The EVOS S1000 Spatial Imaging System allows capturing multiplex immunofluorescence images through its spectral unmixing capabilities.  These spectra show emissions of eight Alexa Fluor and Alexa Fluor Plus dyes and DAPI. Despite the overlap, the built-in algorithms in the EVOS S1000 spatial imaging software can determine the relative contribution of each fluorophore to every pixel of the image and eliminate the spectral bleedthrough from overlapping channels.
Once all these components are collected, the EVOS S1000 software generates and saves the unmixing matrix to the imaging protocol (Figure 5). 
Figure 5. The EVOS S1000 Spatial Imaging System generates multiplexed data through its spectral unmixing capabilities. This software feature allows researchers to easily visualize all channels simultaneously and provides a quality metrics report to facilitate highly resolved data.
Gain transparency and confidence in unmixing quality
An Unmixing Quality Metric Report enables the evaluation of the experimental panel before time is spent imaging full tissue scans (Figure 6). This report provides guidance and metrics demonstrating that the bleed through from spectrally overlapping markers will be removed after the unmixing protocol is applied to spectrally mixed multiplex tissue samples.
Figure 6Visualization of the raw images (left) and unmixed images (right) for each single-color control sample, displayed in each column, across the primary channels shown in each row.
Traditional 1-color staining highlights specific proteins
Figure 7. Stained invasive ductal carcinoma shown in individual tiles. Human invasive ductal carcinoma of breast tissue processed and stained with DAB or hematoxylin. Tiles represent individual targets. Images were taken on the EVOS S1000 Spatial Imaging System.
Multiplex immunohistochemistry staining highlights the complexity of biological systems within tissues
Figure 8Invasive ductal carcinoma tissue stained with the 8-plex Aluora spatial amplification assay and DAPI. Human invasive ductal carcinoma of breast tissue processed and stained with the Aluora Spatial Rainbow Kit (Cat. No. A40002450). Images and spectral unmixing were performed on the EVOS S1000 Spatial Imaging System. Data was analyzed for single cell segmentation and phenotyping to reveal spatial distribution of immune cell subpopulations. Analysis of the multiplex immunofluorescence stitched image was performed on the Indica Labs HALO (version 4.0.5107.318) software.
Simplified tissue imaging workflow
Unlike existing technologies that can take several days to weeks, the EVOS S1000 instrument can complete the imaging process within several hours, with the greatest time savings found in its capability to generate a fully stitched, unmixed, multiplexed image for multiple samples. With the EVOS S1000 Spatial Imaging System, you have the flexibility to use a variety of labeling technologies, allowing you to select and utilize your preferred antibodies and reagents. The instrument outputs OME-TIFF files that are compatible with any third-party analysis software such as Halo and QuPath (QuPath: Open source software for digital pathology image analysis. Scientific Reports (2017).)
Figure 9. The spatial biology multiplex workflow incorporates the EVOS S1000 Spatial Imaging System that multiplexes more biomarkers with the use of spectral unmixing.
To build a panel for the EVOS S1000 Spatial Imaging system, start by choosing your targets, ensuring they align with your research goals. Next, create the panel by carefully picking out antibodies; use the Invitrogen SpectraViewer to determine where to place the 8 fluorophores labeling the antibodies plus DAPI. For the staining and labeling technology, use our Spatial Biology Reagent Selection Tool to chose what is right for your experiment. Finally, proceed with the imaging step to capture and analyze your samples.
Get more information through multiplexed immunofluorescence with a simplified spatial imager that shows the complexity of your tissue
Generate more meaningful images with the EVOS S1000 Spatial Imaging System. Use the OME-TIFF images with many data analysis programs to capture:
  • Cell types, cell states and cell functions
  • Cellular identify in time and space
  • Cell to cell interactions
  • Cellular “neighborhoods” or networks
  • Tissue microenvironments and architecture in their native state
Figure 10. Colon cancer tissue staining of immune markers and DAPI on the EVOS S1000 Spatial Imaging System. Markers include α-SMA Alexa Fluor 420, CD68 Alexa Fluor Plus 488, CD20 Alexa Fluor 514, CD4 Alexa Fluor Plus 555, CD8 Alexa Fluor Plus 594, FoxP3 Alexa Fluor Plus 647, Pan-CK Alexa Fluor 700, Ki-67 Alexa Fluor Plus 750. 
Figure 11. Left panel is a zoom of the colon adenocarcinoma tertiary lymphoid structure (TLS) as circled in Figure 10. Right panel displays cell identities within the TLS after nuclear segmentation and cellular phenotyping in using HALO data analysis software.
Benefits of spectral unmixing technology
Perform imaging faster
Accelerate your experiments with the EVOS S1000 Spatial Imaging System than traditional cyclic technologies. Complete a full round of 9-plex multiplex immunofluorescence faster using advanced software with spectral unmixing.
Reduce the chance for tissue damage
Prevent extensive tissue damage. Iterative staining using bleaching or antibody-removal methods extends experimental time due to lengthy antibody and reagent incubations, and poses risks of epitope loss, tissue degradation, and incomplete fluorophore inactivation across multiple cycles.
Find confidence in your experiments
Gain peace of mind with the EVOS S1000 Spatial Imaging System automated unmixing matrix generation for imaging-stained multiplex tissue samples. The EVOS S1000 software automatically generates and applies an unmixing matrix to spectrally mixed multiplex tissue samples, clearly identifying fluorophore emissions that bleed into neighboring channels.
Autofluorescence removal
Enhance your imaging precision and improve the accuracy of your cell detection with the EVOS S1000 Spatial Imaging System's spectral unmixing technology.
Remove autofluorescence signals from targets of interest using advanced spectral unmixing technology with the EVOS S1000 Spatial Imaging System software.
Benefits of autofluorescence removal
The EVOS S1000 Spatial Imaging System software facilitates the spectral unmixing of tissue autofluorescence, effectively separating highly autofluorescent signals from desired fluorophore staining. Tissue autofluorescence is present in all tissues to varying degrees and is often strongest in channels where fluorophores such as DAPI are imaged. If left in the image data, autofluorescence can interfere with downstream analysis.
See autofluorescence removal by spectral unmixing in action with the EVOS S1000 Spatial Imaging Software. Slide to toggle Raw and Unmixed.
Much like spectral unmixing of fluorophore labels from each other, autofluorescence removal leverages differences in spectral signatures between autofluorescence signals and fluorophore labels. This technology determines the abundance of signals belonging to each signature, allowing for precise separation of highly spectrally overlapping fluorophores.
Improve accuracy of downstream cell detection
Without autofluorescence removal, downstream image analysis becomes more challenging due to uncertainty about whether pixel intensities belong to a desired marker, such as DAPI, which is commonly used for nuclear segmentation, or tissue autofluorescence. By removing the contribution of tissue autofluorescence from DAPI staining, a much higher confidence cell segmentation mask can be obtained using various nuclear segmentation methods.  
Figure 13. Cell detection performed on DAPI without unmixing (top) compared to cell detection performed on DAPI after unmixing and autofluorescence removal (bottom).
EVOS S1000 system–compatible antibodies and reagents
Conjugated primary antibodies
Detect numerous targets simultaneously using conjugated primary antibodies for multiplexing experiments. These antibodies have been tested on tissue samples and are available in formats compatible with the EVOS S1000 Spatial Imaging System and other microscopy platforms.
Aluora Spatial Amplification Kits
Detect up to 8 targets simultaneously in one tissue sample with Aluora Spatial Amplification Kits. These kits combine bright Aluora dyes with enzyme-mediated signal amplification to help overcome autofluorescence in the detection of rare targets.
ReadyLabel Antibody Labeling Kits
Overcome the challenges of assembling multiplex imaging panels by directly conjugating your primary antibody with dye colors optimized for spatial analysis.
ViewRNA ISH Tissue Assay
Detect multiple RNA targets within one tissue sample. The kit provides exceptional sensitivity and allows single-molecule detection of up to four RNA targets simultaneously.
Aluora FluoCells Prepared Slide Controls
Establish reference spectra, set acquisition parameters, and practice spectral unmixing with ready-to-use FFPE murine kidney slides stained with Aluora dyes.
Need help with designing your multiplex imaging experiments? Contact our spatial imaging experts here.
Customer story: SciLifeLab, Stockholm, Sweden, Spatial Proteomics Unit
Using the Invitrogen EVOS S1000 Spatial Imaging System simplifies the tissue imaging process and provides multiplex data in less than one hour.
"With the new EVOS S1000, we can run more projects in parallel. It allows us to achieve multiplexing without needing to remove antibodies or fluorophores and it significantly reducing sample processing time"
- Carolina Osés Sepúlveda, SciLife Stockholm, Sweden
“The EVOS S1000 has a very fast software, I like the live [periscope] mode. I also like the simplicity of the unmixing workflow. It is really compatible with analysis software like QuPath!”
-Giulia Bergamaschi, Amsterdam Universitair Medische Centra (UMC), The Netherlands
"The software is very intuitive and easy to explain. The images are super crisp and it's great to be able to see the scanning progress in real time. The spectral unmixing algorithm really makes signals stand out”
-Maria Lung, SciLife Stockholm, Sweden
Ordering information
Every EVOS S1000 Spatial Imaging System (Cat. No. AMFS1000) includes the imaging unit as well as the objectives (1 of each type), slide holders (4), calibration slide, and AB Assurance support plan listed below. 
Every EVOS S1000 Spatial Imaging System (Cat. No. AMFS1000) includes the imaging unit as well as the objectives (1 of each type), slide holders (4), calibration slide, and AB Assurance support plan listed below. 
Catalog # Name Size Price Qty
B10710 Each
143.65
Online Exclusive
191.00 Save 47.35 (25%)
R3777 Each
267.00
152885000 Each -
P36980 Each
343.65
Online Exclusive
378.00 Save 34.35 (9%)
S36917 Each
87.65
Online Exclusive
94.75 Save 7.10 (7%)
Resources and support
Spatial imaging protocol handbook
Read and use trusted protocols for many types of imaging including 3D staining, multiplexing ISH in tissue, and other spatial biology protocols.
Spatial biology resource center
Find resources including webinars, conference posters, protocols, guides, handbooks, and publications to start your multiplex experiments.
SpectraViewer
Explore the excitation and emission spectra of fluorophores.
Spatial Reagent Finder
The Spatial Biology Reagents Selection Tool helps you find the reagents compatible with your multiplex IHC experimental needs.
Publications
Application notes
Scientific posters
Selection guides
Specification sheet
Other articles
How-to tutorial videos
The Invitrogen EVOS S1000 Spatial Imaging System is available in all participating countries and regions except for Central and South America.
HALO is a trademark of Indica Labs, Inc.
For Research Use Only. Not for use in diagnostic procedures.