1x1 image pixel for data collection
High-performance general purpose centrifuges
Thermo Scientific X Pro General Purpose Centrifuges are designed to provide exceptional performance, power, and versatility through innovations to support efficiency across applications. With fixed-angle rotor RCF (Relative Centrifugal Force) of over 25,000 x g and swinging bucket rotor RCF of over 7,000 x g, even the toughest separations can be completed with confidence.
Scientists working across biopharmaceutical, academic research and clinical diagnostic applications can choose between ventilated or refrigerated 1.6 L (4 x 400 mL) and 4 L (4 x 1,000 mL) floor and bench multi-purpose models.
Key product highlights
Touchscreen interface
Easy programming, health icon, quick spins as well as useful information and support are provided on the full-color touchscreen. Monitor your runs with the lighthouse mode and save time with the pre-temp button.
x pro touchscreen
Auto-Lock Rotor Exchange
Ergonomic push-button rotor exchange can help save time. In as little as three seconds, you can change rotors, or remove them for cleaning purposes. No tools needed.
auto-lock rotor exchange
ClickSeal Biocontainment Lids
Certified*, secure and quick solution to help ensure that when used correctly, pathogens can be safely contained in the event of a tube breakage, tube leakage, external tube contamination or over-fill contamination within tube and cap threads.
clickseal biocontainment lids
Available models and centrifuge selection
X1 Pro Benchtop Centrifuges
X4 Pro Benchtop Centrifuges
X4F Pro Floor Model Centrifuges
Description
Up to 1.6L capacity
Benchtop ventilated (X1)
or
refrigerated (X1R) centrifuge models
Up to 17 rotor options
Up to 4L capacity
Benchtop ventilated (X4)
or
refrigerated (X4R) centrifuge models
Up to 18 rotor options
Up to 4L capacity
Floor-standing ventilated (X4F)
or
refrigerated (X4FR) centrifuge models
Up to 18 rotor options
Models/Brands
Sorvall X1 and X1R Pro Centrifuges
Sorvall X4 and X4R Pro Centrifuges
Sorvall X4F and X4FR Pro Centrifuges
Multifuge X1 and X1R Pro Centrifuges
Multifuge X4 and X4R Pro Centrifuges
Multifuge X4F and X4FR Pro Centrifuges
Centrifuge features and benefits
x-pro features infographic
User interface Wide range of rotors
Modern, intuitive touchscreen interface offers easy access, programming and monitoring of the centrifuge runs. The interface provides up to 100 run programs, a pre-temp button, a customizable pulse button for quick on-the-fly spins, scheduling features and preventative maintenance queues.  Up to 18 rotor options for our 4 L centrifuges and up to 17 rotors for our 1.6 L models are available to accommodate a variety of sample types and vessels. Select up to 7 Fiberlite Carbon Fiber Rotors (up to 5 Fiberlite options with the smaller models) with their lightweight, durable design and a 15-year limited warranty.
Auto-Lock rotor exchange Energy efficiency and sustainability
Quick rotor exchange technology helps provide the flexibility to quickly swap between your sample preparation protocols. The easy and reliable push-button system replaces the need for tools to bolt down the rotor, shortening run set-up time.
Download Auto-Lock technology Smart Note 
Scheduling features and carbon fiber rotors help save energy. The X Pro centrifuges are RoHS and WEEE compliant and utilize recyclable packaging materials. They are manufactured at a certified green energy site (Osterode, Germany).
Safety, ergonomics and convenience Connectivity
For enhanced ergonomics, the loading height and the lid gripper height decreased and there is less effort required to close the lid due to motorized gas springs compared to previous models. Smart technologies like the SmartSpin imbalance detection and Accumulated Centrifugal Effect (ACE) function provide additional convenience and help support safety.
With the optional Secure-Spin Mode, the X Pro centrifuges allow data collection, reporting, and archiving, as well as management and protection of process data for 21 CFR Part 11 compliance. Once your centrifuge has the Secure-Spin Mode it is also compatible with Centri-Log Plus Data Management Software.
Biocontainment lids  Variety of certified and dedicated models
Glove-friendly sealing option for one-handed operation and providing audible and visual closure cues. Available for multiple swing-out and fixed angle rotors. The biocontainment of our ClickSeal lids is third-party certified*.
Download ClickSeal Lids Smart Note
Choose between general use or IVD. Benefit from cleanroom compatibility of our 4 L models and the Cell Therapy Systems Series models with documentation.
*Certified by the National Institute for Health Protection, Porton Down, UK or TÜV NORD CERT GmbH, Germany.
Packages
The GP X Pro centrifuges are available as packages including centrifuge, rotor cross, buckets, lids and adapters for cell culture or blood tube applications.
centrifuge h-flex package 800
Cell culture packages
  • Cell culture package (TX-400): GP X1 Pro 1.6 L centrifuge (ventilated or refrigerated) + TX-400 rotor, buckets, biocontainment lids and adapters for 36 x 15 mL and 16 x 50 mL conical tubes.
  • Cell culture package (H-Flex): GP X1 Pro 1.6 L centrifuge (ventilated or refrigerated) + H-Flex rotor, buckets, biocontainment lids and adapters for 36 x 15 mL and 16 x 50 mL conical tubes. (Only available in North America)
  • Cell culture package (TX-750): GP X4 Pro 4 L centrifuge (ventilated or refrigerated) + TX-750 rotor, buckets, ClickSeal lids and adapters for 56 x 15 mL conical tubes and 28 x 50 mL conical tubes.
  • Cell culture package (TX-1000): GP X4 Pro 4 L centrifuge (ventilated or refrigerated) + TX-1000 rotor, buckets, ClickSeal lids and adapters for 96 x 15 mL conical tubes and 40 x 50 mL conical tubes.
Blood tube packages
  • Blood tube package (TX-400): GP X1 Pro 1.6 L centrifuge (ventilated or refrigerated) + TX-400 rotor, buckets, biocontainment lids and adapters for 76 x 5/7 mL blood tubes and 56 x 10 mL blood tubes.
  • Blood tube package (TX-750): GP X4 Pro 4 L centrifuge (ventilated or refrigerated) + TX-750 rotor, buckets, ClickSeal lids, adapters for 148 x 5/7 mL blood tubes and 84 x 9/10 mL blood tubes.
  • Blood tube package (TX-1000): GP X4 Pro 4 L centrifuge (ventilated or refrigerated) + TX-1000 rotor, buckets, ClickSeal lids and adapters for 196 x 5/7 mL and 148 x 10 mL blood tubes.
Additionally, we offer rotor packages, where you can buy the centrifuge separately and then the rotor with necessary accessories for your application.
Rotor packages (purchase the centrifuge separately)
  • H-Flex rotor packages (for 1.6 L models only): Tissue Culture Package | Blood Tube Package | Microplate Package.
  • TX rotor packages (TX-400, TX-750, TX-1000): View all
Cell Therapy Systems Series
For cell and gene therapy workflows, we offer the X4 Pro Centrifuge CTS (Cell Therapy Systems) Series which consists of the product and a comprehensive documentation package including certifications and protocols. Compliance services are available to support a quick implementation into processes for GMP needs.
X4 pro CTS series 800px
Certifications
  • Choose between general use or IVD (clinical) models. Our clinical models are in compliance with the EU IVDR (EU) 2017/746 and are listed by the FDA.
  • Available with the following certifications: UL, CE, RoHS, CSA.
  • Standards: IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, EN ISO 13485.
  • Cleanroom compatibility of 4L models: Class ISO 7 according to DIN EN ISO 14644-1.
Made in Germany
  • Our X Pro centrifuges are developed and manufactured in Osterode am Harz, Germany. For over 75 years, our center of excellence in Osterode has been at the forefront of designing and manufacturing innovative centrifuges. We take pride in partnering with our customers to develop efficient and intuitive products that help meet their specific needs. 
Watch the video below to learn about our site
Video Player is loading.
Current Time 0:00
Duration 2:30
Loaded: 0%
Stream Type LIVE
Remaining Time 2:30
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    3D virtual tour
    3D virtual tour of X4R Pro Centrifuge
    360-degree, spin, zoom, rotate, discover and interact with our virtual X Pro Series.
    Resources
    Video
    Thermo Scientific X Pro centrifuges feature improved reach-in for loading, a touchscreen interface, single-screen programming, and more.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.