Peroxisome labeling using SelectFX® Alexa Fluor® 488 Peroxisome Labeling Kit, for fixed cells:
Peroxisomes were labeled using an antibody directed at peroxisomal membrane protein 70 (PMP-70) and detected with Alexa Fluor® 488 dye–labeled goat anti–mouse IgG secondary antibody. Mitochondria were stained with MitoTracker® Red CMXRos (Cat. no. M7512) prior to fixation; nuclei were stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490).
Peroxisomes were labeled using an antibody directed at peroxisomal membrane protein 70 (PMP-70) and detected with Alexa Fluor® 488 dye–labeled goat anti–mouse IgG secondary antibody. Mitochondria were stained with MitoTracker® Red CMXRos (Cat. no. M7512) prior to fixation; nuclei were stained with blue-fluorescent DAPI (Cat. no. D1306, D3571, D21490).
Related Products
Catalog # Name Size Price (USD) Qty
D1306 DAPI (4',6-diamidino-2-phenylindole, dihydrochloride), 10 mg, 10 mg Each
152.65
Online Exclusive
156.00
Save 3.35 (2%)
D21490 DAPI, FluoroPure™ grade, 10 mg, 10 mg Each
225.65
Online Exclusive
231.00
Save 5.35 (2%)
D3571 DAPI (4',6-diamidino-2-phenylindole, dilactate), 10 mg, 10 mg Each
151.65
Online Exclusive
156.00
Save 4.35 (3%)
M7512 MitoTracker™ Red CMXRos - Special Packaging, 20 x 50 μg, 20 x 50 μg Each
506.65
Online Exclusive
522.00
Save 15.35 (3%)
Related Images