KingFisher™ Presto Purification System
Thermo Scientific™
KingFisher™ Presto Purification System
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeVolume (Metric) Processing
5400830KF Presto with 96 DW head50 to 1000 μL
2407883096 DW head50 to 1000 μL
2407883196 DW head and heating block50 to 1000 μL
2407884024 Deep Well head200 to 5000 μL
2407884124 DW head and heating block200 to 5000 μL
5400840KF Presto with 24 DW head200 to 5000 μL
Catalog number 5400830
Price (USD)
-
Product Type:
KF Presto with 96 DW head
Volume (Metric) Processing:
50 to 1000 μL
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories. Utilizing proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Features:
• Purify 96 or 24 samples at a time
• Volume range of 50–5000 μL
• Minimized size for easy fitting to automation systems
• Heating capability for application requiring elevated temperatures
• Easy connection to several liquid handling instruments
• Flexible system for wide variety of applications
Specifications
Depth (English)18.3 in.
Depth (Metric)465 mm
Heating TemperatureFrom +5°C above ambient temperature to 115°C
Height (English)15.7 in.
Height (Metric)400 mm
Plate Type96, 96 DW
Samples per Run96
Weight (English)52.9 lb.
Weight (Metric)24 kg
Width (English)14.2 in.
Width (Metric)36 cm
Product TypeKF Presto with 96 DW head
Volume (Metric) Processing50 to 1000 μL
Unit SizeEach
Frequently asked questions (FAQs)
What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?
If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.