Your bioprocessing workflow, reinvented
Labtainer Pro BPC with BioTitan Retention Device
The Labtainer BPC Series is designed to deliver a complete range of 2D containers engineered to match the specific needs of each workflow stage. Whether you require accessible, everyday performance or the highest level of quality assurance, Labtainer BPCs offers  the right combination of design, materials, and manufacturing rigor to support reliable, efficient operations.
Key features of the Labtainer BPC Series
Essential performance for everyday bioprocessingBioProcess Container in-process inspection methods
  • Proven, dependable design for routine upstream and general processing applications
  • Broad selection of robust film options, including trusted Aegis5-14 and CX5-14 films
  • BioTitan Retention Device for secure, reliable port connections
  • Streamlined configurations that deliver value and compatibility across common workflows
  • In-process visual and automated inspections to support consistent product quality
Enhanced design and usability for greater workflow efficiency 
Upgraded Labtainer Pro BPC outer packaging
  • Optimized chamber drainage reduces holdup and supports efficient liquid evacuation with less manipulation
  • Improved port sealing with impulse heat–sealing method to support strong port welds and consistent seal performance during filling, handling, and transfer
  • Ergonomic handling features help operators lift, carry, and position the BPC more easily in routine use
  • Configurable port layouts minimize unused ports and help reduce potential failure points
  • Offered in robust Aegis5-14 and CX5-14 films, produced in controlled cleanroom environments to help support consistent film properties and durability
  • Well suited for applications that require elevated efficiency and handling performance compared to standard BPCs
Enhanced quality controls for higher-assurance applicationsLabtainer Pro BPC improved handle feature
  • 100% helium integrity testing is performed on all standard configurations to support confidence in BPC containment performance
  • Lot-based USP endotoxin (USP <85>) and particulate testing (USP <788>) provide additional verification of product quality for applications needing defined testing requirements
  • Constructed using robust Aegis5-14 and CX5-14 films, manufactured in controlled cleanroom environments to support film consistency and strength
  • Manufactured under precise, documented process controls designed to help maintain reproducibility across production lots
  • Well suited for downstream, filling, or regulatory-driven processes where added quality verification steps are required
Essential performance for everyday bioprocessingBioProcess Container in-process inspection methods
  • Proven, dependable design for routine upstream and general processing applications
  • Broad selection of robust film options, including trusted Aegis5-14 and CX5-14 films
  • BioTitan Retention Device for secure, reliable port connections
  • Streamlined configurations that deliver value and compatibility across common workflows
  • In-process visual and automated inspections to support consistent product quality
Enhanced design and usability for greater workflow efficiency 
Upgraded Labtainer Pro BPC outer packaging
  • Optimized chamber drainage reduces holdup and supports efficient liquid evacuation with less manipulation
  • Improved port sealing with impulse heat–sealing method to support strong port welds and consistent seal performance during filling, handling, and transfer
  • Ergonomic handling features help operators lift, carry, and position the BPC more easily in routine use
  • Configurable port layouts minimize unused ports and help reduce potential failure points
  • Offered in robust Aegis5-14 and CX5-14 films, produced in controlled cleanroom environments to help support consistent film properties and durability
  • Well suited for applications that require elevated efficiency and handling performance compared to standard BPCs
Enhanced quality controls for higher-assurance applicationsLabtainer Pro BPC improved handle feature
  • 100% helium integrity testing is performed on all standard configurations to support confidence in BPC containment performance
  • Lot-based USP endotoxin (USP <85>) and particulate testing (USP <788>) provide additional verification of product quality for applications needing defined testing requirements
  • Constructed using robust Aegis5-14 and CX5-14 films, manufactured in controlled cleanroom environments to support film consistency and strength
  • Manufactured under precise, documented process controls designed to help maintain reproducibility across production lots
  • Well suited for downstream, filling, or regulatory-driven processes where added quality verification steps are required
BioTitan retention device
BioTitan device offers 360° of universal retention 
This industry-leading product is also now manufactured with BioTitan Retention Device—for heightened performance and lower risk. The BioTitan device is designed to provide superior universal retention that minimizes the risk of leaks and failures at the connection point. The innovative design concept of the Thermo Scientific Labtainer Pro BioProcess Container (BPC) provides flexibility and assurance without compromise
Request information   Download data sheet
Available accessories
Labtainer Pro BPC Accessories 
The Labtainer BPC series have been engineered with multiple product accessories to help maximize productivity and to help improve the overall experience of working with the product
The Labtainer tote offers BPC protection and access to lines without the need to open the lid. The tray inside the tote is designed with a tilted plate to allow for optimal drainage. This product is constructed of medical-grade, high-density PE.
Labtainer Pro BPC Interactive
Using this interactive tool, browse the key features and benefits of the Labtainer Pro BPC. Interact with the fill and dispense feature while seeing a 360° view of all available sizes ranging from 50 mL to 20 L.
Labtainer Pro BPC Interactive Tour
Ordering information
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Explore our resources
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.