Lentiviral vector production capabilities
Suspension solutions
The Gibco LV-MAX Lentiviral Production system is the first optimized system that provides all the high-quality components you need in a chemically defined, serum-free environment.
  • Cost-effective—reduce cost by more than 50%**
  • High titers—greater than 1 x 108 TU/mL (unconcentrated)
  • Research-grade and CTS options—seamlessly transition from discovery to commercial production
Request information
The Gibco LV-MAX Lentiviral Production System includes: HEK 293–derived suspension cells, serum-free media, proprietary transfection reagent, supplement, and our novel enhancer. Our HEK 293 derived viral production cells have been optimized for viral production in suspension culture.
Optimized and fully integrated system
Illustration of the seven stages from suspension culture to LV particles
Figure 1. LV-MAX Lentiviral Production System protocol overview.
Multi-component system optimized for improved lentivirus production
The LV-MAX Lentiviral Production System provides a scalable and high-yield lentiviral vector production platform. It is based on a high-density suspension culture of HEK 293–derived viral production cells in LV-MAX Production Medium. Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel lentivirus-specific enhancer and supplement (Figure 1). All components work synergistically to help generate superior, functional lentiviral particles under xeno-free conditions compared to conventional polyethylenimine (PEI), serum-based culture systems (Figure 2).
Bar chart demonstrating lentivirus titers are dramatically improved using LV-MAX Lentiviral production system
Figure 2. Significantly higher viral titers with LV-MAX system.
These data highlight the positive impact of using our complete system to maximize production of lentiviral vectors compared to titers obtained when using incomplete or non-optimized systems (Figure 3). Lentivirus was produced in 30 mL shaker flasks using our LV-MAX Lentiviral Production System or alternative transfection reagents and cells. LV293 = Gibco viral production cells (HEK 293–derived suspension cells), 293F = Gibco FreeStyle 293-F cell line, PEI = polyethylenimine.
High titers
The LV-MAX system produced up to 15 times more virus than the PEI-mediated system in adherent cells and up to 10x more virus than in suspension cells (Figure 3), resulting in over 50% cost reduction compared to polyethylenimine (PEI)–based lentiviral production methods.
Lentivirus production comparison for optimized kit versus PEI competitor
Figure 3. Lentivirus titers are dramatically improved using LV-MAX Lentiviral Production System.
Cost effective
Unfiltered lentivirus produced by suspension cells using the LV-MAX Lentiviral Production System was compared with polyethylenimine (PEI)–mediated transfection of lentiviral vectors in adherent HEK 293T/FT cells and suspension HEK 293 cells. The lentiviral titer was determined by transducing HT1080 cells and analyzing GFP-positive cells. The resulting cost savings is also shown.
Current production methodSwitch to LV-MAX system and save*
Adherent PEI58%
Suspension PEI52%
* Cost comparison based on list price in USD and LV yield of 1 x 108 TU/mL using LV-MAX and 1 x 107 TU/mL using PEI-based adherent and suspension methods. Cost consideration includes media, transfection reagents, and culture vessels.
Ordering information for LV-MAX kits and components
GMP cells and media
Catalog # Name Size Price (USD) Qty
A3583402 LV-MAX™ Production Medium, 6 x 1L Each
1,764.65
Online Exclusive
1,914.00
Save 149.35 (8%)
A35347 Viral Production Cells, 1 mL Each
1,306.00
K497500 ViraPower™ Lentiviral Packaging Mix Each
3,845.00
A35348 LV-MAX™ Transfection Kit Each
1,650.65
Online Exclusive
1,786.00
Save 135.35 (8%)
31985088 Opti-MEM™ I Reduced Serum Medium, 10 x 500 mL Each
613.65
Online Exclusive
676.00
Save 62.35 (9%)
A35827 Viral Production Cells, 6 x 1 mL Each
7,010.00
A43237 LV-MAX™ Lentiviral Packaging Mix Each
696.65
Online Exclusive
740.00
Save 43.35 (6%)
31985062 Opti-MEM™ I Reduced Serum Medium, 100 mL Each
31.65
Online Exclusive
34.34
Save 2.69 (8%)
A35684 LV-MAX™ Lentiviral Production System Each
2,405.00
31985070 Opti-MEM™ I Reduced Serum Medium, 500 mL Each
72.65
Online Exclusive
78.75
Save 6.10 (8%)
A3583401 LV-MAX™ Production Medium, 1 L Each
317.65
Online Exclusive
340.00
Save 22.35 (7%)
Advanced lipid nanoparticle technology for superior lentiviral production in adherent cultures
Invitrogen Lipofectamine 3000 Transfection Reagent is a highly efficient, cost-effective tool for lentiviral production. This versatile reagent enables high viral titers even with genes that are large or difficult to package.
Invitrogen Lipofectamine 3000 Reagent
  • High titers—including genes that are large or difficult to package
  • Gentle—reduced reagent dose for improved cell viability
  • Flexible—compatible with your existing protocol
  • Cost-effective—use less ancillary components and labor
Invitrogen Lipofectamine 3000 reagent
In a side-by-side comparison between Lipofectamine 3000 Reagent, Lipofectamine 2000 Reagent and PEI, Lipofectamine 3000 reagent–mediated lentiviral production was ~5–10 times greater than that obtained from the other transfection reagents (Figure 5).
bar chart showing lentivirus production in adherent cells with Lipofectamine 3000 reagent
Figure 5. Lentivirus production in adherent cells with Lipofectamine 3000 reagent. pLenti6.3/V5-GW/EmGFP and Invitrogen ViraPower Lentiviral Packaging Mix were delivered by Lipofectamine 3000 reagent, Lipofectamine 2000 reagent, and polyethylenimine (PEI) in a 6-well culture plate with HEK 293T/FT cells. The lentiviral titer was determined by transducing HT1080 cells and analyzing GFP-positive cells.
Ordering information for Lipofectamine 3000 Reagent
Catalog # Name Size Price (USD) Qty
L3000015 Lipofectamine™ 3000 Transfection Reagent, 1.5 mL Each
877.65
Online Exclusive
938.00
Save 60.35 (6%)
L3000001 Lipofectamine™ 3000 Transfection Reagent, 0.1 mL Each
89.65
Online Exclusive
94.00
Save 4.35 (5%)
L3000008 Lipofectamine™ 3000 Transfection Reagent, 0.75 mL Each
563.65
Online Exclusive
598.00
Save 34.35 (6%)
L3000150 Lipofectamine™ 3000 Transfection Reagent, 15 mL, 15 mL Each
7,814.65
Online Exclusive
8,140.00
Save 325.35 (4%)
L3000075 Lipofectamine™ 3000 Transfection Reagent, 5 x 1.5 mL Each
4,222.65
Online Exclusive
4,550.00
Save 327.35 (7%)
Transfection
Is my CTS LV-MAX transfection reagent supposed to be cloudy?
It is normal to see some turbidity and cloudiness in LV-MAX transfection reagent. Lipid transfection reagents are sensitive to low temperature; if you store them at temperatures lower than the recommended 2–8°C storage conditions or if the reagent freezes it will precipitate, leading to low transfection efficiency or inactivity.
I'm interested only in the transfection reagent. Can I purchase it separately from the transfection kit?
Not currently. The LV-MAX Transfection Kit comes with three components that are not sold a la carte. The system components were specifically optimized to work as a system for maximal viral vector titer. We can however customize different ratios of the supplement, enhancer and transfection reagent.
Have you tried other commercially available transfection reagents (i.e., PEI) in your system?
Yes, we have tried a full range of transfection reagent formulations while developing the system. LV-MAX Transfection Reagent is compatible with the suspension Viral Production Cells to support transfection in a high cell density format and offers higher performance than other transfection reagents.
LV-MAX is more expensive than PEI-based transfection reagent. How can you claim it’s a low-cost option?
The cost per viral particle is much lower using LV-MAX system compared to PEI-based method due to the robustness of the LV-MAX system. Because of the high efficiency of the complete system, less material and volumes are needed to make the same amount of virus, which also reduces labor and ancillary consumable reagents (plasmids, purification etc.).
Can I transfect before passage 5 (P5)?
We recommend that cells be used for lentiviral production following passage 5 to allow for full recovery of the cells from the cryopreservation.
Enhancer addition time is suggested to be anytime between 5–14 hours after transfection, how critical is it that we stay within this time frame window?
Enhancer addition was tested over the entire rage of 5–14 hours and was found to perform with equivalent efficiency when added anytime during that range of time. The enhancer formulation coupled with the broad window of addition time was specifically optimized to allow the user flexibility in schedule and start time of the assay without affecting performance.
Production
What comes with the CTS Viral Production Cells?
We have extensive documentation to demonstrate lineage history, viral clearance, and traceability of our GMP manufactured process. Under CDA, we can share our Certificate of Analysis and Table of Contents. Full access to documentation package is available through outlicensing@thermofisher.com.
Can I use the CTS Viral Production Cells or CTS LV-MAX system for clinical trials or commercial use?
Please refer to our standard sales terms and conditions and limited use label license for specific products. For further inquiries, please contact us at outlicensing@thermofisher.com.
My cells are initially low viability post thaw, is this normal?
The protocol recommends culturing the cells to passage 5 (P5) so that the cells recover sufficiently prior to starting the protocol for LV production. It is normal to see variance in viability upon thaw, but this should normalize during recovery passages.
What density should I be seeding the cells?
Depending on whether you are doing a 3 day or 4-day culture, we have outlined the differences in seeding densities in our LV-MAX user guide manual. We also have specific guidance around seeding densities to be used for lentiviral vector production.
Why do we have to dilute the cells once more before transfection? Is that necessary?
This is a necessary step to replenish nutrients to the cells and ensure that the cells are at the correct density to achieve the concentrations needed for optimal transfection efficiency. Proper cell density at time of transfection ensures maximal performance of the system.
Do you have recommendations for how to measure non-fluorescent LV titer?
In our user guide manual, we provide a protocol for titer measurement using antibiotic selection.
Why is there no media exchange requirement in the LV-MAX protocol and the cells remain in the same media throughout the two-day transfection? What data do you have to check on the effect of media exchange?
We have done comprehensive studies on the media changes. The LV-MAX production medium is specially designed to support the health of the Viral Production Cells cultured at high-density. Therefore, there’s no need for media exchanges.
The Viral Production Cells that come with the starter LV-MAX Lentiviral Production System are supposed to be good in passage 5–20. Does this refer to passages 5–20 post-thaw, or should we consider the number of passages the cells went through before cryopreservation?
The passages should be counted upon thaw and discarded when they reach passage 20 post-thaw.
Should I harvest at 24 hours and 48 hours? What about waiting for 72 hours? What’s the best harvest method?
Our system has been optimized to provide maximal lentiviral production at 48 hours post transfection. Waiting until 72 hours is not recommended since existing particles within the media will infect the production cells and can reduce the titer.
Commercialization
What is the intended claim for these products?
LV-MAX products are intended for research use only while our CTS LV-MAX products are intended for research use or manufacture of cell, gene or tissue-based products. Please refer to individual product for further details on the intended use claim.
What are the major differences between LV-MAX and the CTS LV-MAX products?
Performance between the two systems is equivalent. In contrast to the research use only LV-MAX products, CTS LV-MAX products have an additional intended use for cell and gene therapy applications and complimented with a more robust regulatory support package to enable clinical therapeutic applications. This means that to the best of our ability, we keep our CTS product specifications and release requirements up to date in accordance with the latest regulatory guidance specific for this intended use.
Do I need to obtain licensing rights if I just want to try the system?
Please refer to our standard sales terms and conditions and limited use label license for specific products. For additional information, please contact us at outlicensing@thermofisher.com.
Have you tried other media like HEK 293 expression media with your system?
Yes, we have tried other media and have seen reduced performance. For maximal LV vector yield, we recommend using LV-MAX medium as part of the entire LV-MAX system.
What scale have you tested for scale up so far? Do you have protocols you can share?
We continually develop large scale protocols to support clinical and commercial LV vector production. Please contact cellculturesupport@thermofisher.com  or the latest information.
What is the recommended ratio of plasmids to use in the system?
We recommend using the lentiviral packaging plasmids to the transfer plasmids at a ratio of 3:2.
Have you tried other plasmids with your systems?
The LV-MAX production system is designed to be compatible with any format of plasmid DNA. The quality of plasmids (e.g., purity), ratio of the plasmids, and vector design all contribute to viral titer. Please contact us at cellculturesupport@thermofisher.com for suggestions.
I need support in designing my plasmid constructs. Who should I reach out to?
Please contact our GeneArt department for assistance at geneartsupport@thermofisher.com.
Looking for a partner to help accelerate your discovery?
Our dedicated staff of R&D scientists and project managers are ready to support you with end-to-end services for every step in your lentivirus (LV) RUO production. Learn more about our viral vector production services.
Support
Support Center
  • Lentiviral Gene Delivery for Mammalian Expression Support
    Explore our resources, tip and tricks, and troubleshooting advice for delivering your lentiviral construct into mammalian cells for expression of your protein of interest. Obtain detailed information on every step of the workflow, including preparation of the lentiviral construct, generation of the lentiviral stock, viral titering, transduction, and analysis of expression.
Technical questions?
** Compared with PEI-based lentiviral production methods.
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.