Micrococcus luteus and Salmonella oranienburg.
Live Micrococcus luteus and Salmonella oranienburg bacteria stained with the LIVE BacLight™ Bacterial Gram Stain Kit (Cat. No. L7005). Gram-positive M. luteus cells fluoresce orange, whereas gram-negative S. oranienburg cells fluoresce green. This image was photographed in a single exposure through an Omega Optical triple bandpass filter set.
Live <i>Micrococcus luteus</i> and <i>Salmonella oranienburg</i> bacteria stained with the LIVE <i>Bac</i>Light™ Bacterial Gram Stain Kit (Cat. No. L7005). Gram-positive <i>M. luteus</i> cells fluoresce orange, whereas gram-negative <i>S. oranienburg</i> cells fluoresce green. This image was photographed in a single exposure through an Omega Optical triple bandpass filter set.
Related Products
Catalog # Name Size Price (USD) Qty
L7005 LIVE BacLight™ Bacterial Gram Stain Kit, for microscopy & quantitative assays Each
607.65
Online Exclusive
626.00
Save 18.35 (3%)
Related Images