eBioscience™ ELISA/ELISPOT Diluent (5X)
Product Image
Invitrogen™
eBioscience™ ELISA/ELISPOT Diluent (5X)
The 5X ELISA/ELISPOT Diluent is a Phosphate Buffered Solution supplemented with fetal bovine serum as a source of animal serumRead more
Have Questions?
Catalog NumberQuantity
00-4202-56100 mL
Catalog number 00-4202-56
Price (USD)
101.65
Online Exclusive
109.00
Save 7.35 (7%)
Each
Add to cart
Quantity:
100 mL
Price (USD)
101.65
Online Exclusive
109.00
Save 7.35 (7%)
Each
Add to cart
The 5X ELISA/ELISPOT Diluent is a Phosphate Buffered Solution supplemented with fetal bovine serum as a source of animal serum proteins. This buffer is used for blocking non-specific binding of ELISA plates, diluting recombinant standards and test samples, and making the working dilutions of the secondary detection reagent (i.e., detection antibody-biotin and AV-HRP Cat. No. 18-4100). This diluent solution also contains a preservative.
Reported Application
ELISA
For Research Use Only
Specifications
For Use With (Application)ELISA
FormLiquid
Product LineeBioscience™
Product TypeDiluent
Quantity100 mL
Unit SizeEach
Frequently asked questions (FAQs)
What other reagents will I need to run an ELISA using the antibody pairs?
When working with our matched pairs or uncoated ELISA kits, please refer to the user guide provided with your antibody-matched pair for kit-specific guidance. When designing your own ELISA, please utilize our ELISA Builder tool for reagent selection as well as our ELISA Development and Optimization pages for futher guidance.
Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link