Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Equipment) | IonPac AS22 Columns |
Type | Eluent Concentrate |
Standard Concentration | 0.45, 0.14 M |
Concentration | 100 X |
Hazards | Irritant |
Standard Components | Sodium Carbonate, Sodium Bicarbonate in Deionized Water |
Quantity | 250 mL |
Description | AS22 Sodium Carbonate/Bicarbonate Concentrate; 100 X; 250 mL |
Storage | Room Temperature |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Standard Components | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
063965 | Each 1 | Sodium Carbonate, Sodium Bicarbonate in Deionized Water | 116.00 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|