Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™
TrackIt™ Ultra Low Range DNA Ladder
Invitrogen TrackIt Ultra Low Range DNA Ladder is ready to use and designed for sizing and approximate quantification of double-strandedRead more
Have Questions?
Catalog number 10488023
Price (USD)/ 50 µg
180.65
Online exclusive
188.00
Save 7.35 (4%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 50 µg
180.65
Online exclusive
188.00
Save 7.35 (4%)
Add to cart
TrackIt™ Ultra Low Range DNA Ladder
Catalog number10488023
Price (USD)/ 50 µg
180.65
Online exclusive
188.00
Save 7.35 (4%)
-
Add to cart
Invitrogen TrackIt Ultra Low Range DNA Ladder is ready to use and designed for sizing and approximate quantification of double-stranded DNA in the range of 10 bp to 300 bp. TrackIt Ultra Low Range DNA Ladder consists of nine individual chromatography-purified DNA fragments and has a reference band at 100 bp for easy orientation.
TrackIt Ultra Low Range DNA Ladder is ideal for separation on 4–5% agarose gels.
Highlights of TrackIt Ultra Low Range DNA Ladder: • Sharp, clear bands—chromatography purified fragments for consistent and reliable results, stable at room temperature up to six months • Ready to use—premixed with 6X TrackIt Cyan/Yellow Loading Dye for tracking of DNA migration • Precise—an exact amount of DNA in each band
Product use The double-stranded DNA ladder can be visualized on 4-5% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration0.1 μg/μL
Gel TypeAgarose
Product TypeDNA Ladder
Ready to LoadYes
Sample Loading Volume1 mL
Shelf Life6 Months
Size Range10 to 300 bp
Volume (Metric)500 μL
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
No. of Reactions100 Applications
Product LineTrackIt™
Quantity50 μg
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TechnologyIndividual chromatography-purified DNA fragments
Unit Size50 µg
Contents & Storage
• 500 μL TrackIt Ultra Low Range DNA Ladder • 1 mL TrackIt Cyan/Yellow Loding Dye
Store at room temperature or at 4°C for up to 6 months. For longer periods store at -20°C.