Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SuperScript™ First-Strand Synthesis System for RT-PCR

Catalog number: 11904018
Have Questions?
Invitrogen™

SuperScript™ First-Strand Synthesis System for RT-PCR

Catalog number: 11904018
Have Questions?
Catalog Number
11904018
Unit Size
50 rxns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1190401850 rxns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The SuperScript First-Strand Synthesis System for RT-PCR is optimized for the synthesis of first-strand cDNA from purified poly(A)+ or total RNA. The system can be used with as little as 1 ng or as much as 5 μg of total RNA. After synthesis, the cDNA can be amplified with specific primers by PCR without intermediate organic extractions or ethanol precipitations. In conjunction with PCR, the system can be used to detect the presence of rare messages, to quantitate the amount of specific mRNA from small numbers of cells, or to clone specific cDNAs without constructing an entire cDNA library. The system is flexible, allowing the use of any PCR enzyme. Combine with AccuPrime Taq DNA Polymerase or Platinum Taq DNA Polymerase for higher specificity PCR or with AccuPrime Pfx DNA Polymerase for high-fidelity cloning applications.

SuperScript II RT
The first-strand cDNA synthesis reaction is catalyzed by SuperScript II Reverse Transcriptase (RT). This enzyme has been engineered to reduce the RNase H activity that degrades mRNA during the first-strand reaction, resulting in greater full-length cDNA synthesis and higher yields of first-strand cDNA than obtained with RNase H+ RTs. Because SuperScript II RT is not inhibited significantly by ribosomal and transfer RNA, it may be used effectively to synthesize first-strand cDNA from a total RNA preparation. The enzyme exhibits increased thermal stability and may be used at temperatures up to 50°C.

Using the SuperScript First-Strand Synthesis System for RT-PCR
This system has been optimized to synthesize first-strand cDNA from varying amounts of starting material. The SuperScript II RT concentration has been lowered and RNaseOUT Recombinant RNase Inhibitor has been added to the system as part of this optimization process. Additionally, reaction conditions have been modified to further increase the sensitivity of the system. Using the kit, you synthesize first-strand cDNA using either total RNA or poly(A)+ selected RNA primed with oligo(dT), random primers, or a gene-specific primer. Then perform PCR in a separate tube using primers specific for the gene of interest.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
GC-Rich PCR Performance
High
Reaction Speed
50 min.
Technique
Reverse Transcription
Optimal Reaction Temperature
42°C
Primer
Random Primers, Oligo dT Primers
Reverse Transcriptase
SuperScript II
Shipping Condition
Wet Ice
For Use With (Application)
RT-PCR
Final Product Type
First-Strand cDNA
No. of Reactions
50 Reactions
Reaction Format
Separate components
Reagent Type
Reverse Transcription
Size (Final Product)
Up to 12 kb
Starting Material
RNA

Contents & Storage

• Oligo(dT)12-18, 50 μL (0.5 μg/μL)
• Random hexamers, 250 μL (50 ng/μL)
• 10X RT buffer, 1 mL
• Magnesium chloride, 500 μL (25 mM)
• DTT, 250 μL (0.1 M)
• dNTP mix, 250 μL (10 mM)
• SuperScript™ II RT, 50 μL (50 U/μL)
• RNaseOUT™, 100 μL (40 U/μL)
E. coli RNase H, 50 μL (2 U/μL)
• DEPC-treated water, 1.2 mL
• Control RNA, 15 μL (50 ng/μL)
• Control Primer A, 20 μL (10 μM)
• Control Primer B, 20 μL (10 μM)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text