1x1 image pixel for data collection
QuantaBlu™ NS/K Fluorogenic Substrate Kit
Product Image
Thermo Scientific™

QuantaBlu™ NS/K Fluorogenic Substrate Kit

The Thermo Scientific QuantaBlu NS/K Fluorogenic Substrate Kit is a two-component kit containing proprietary fluorescent substrate and peroxide solutions forRead more
Have Questions?
Catalog NumberQuantity
15162280 mL
Catalog number 15162
Price (USD)
299.00
Each
Add to cart
Quantity:
280 mL
Request bulk or custom format
Price (USD)
299.00
Each
Add to cart
The Thermo Scientific QuantaBlu NS/K Fluorogenic Substrate Kit is a two-component kit containing proprietary fluorescent substrate and peroxide solutions for non-stopped kinetic assays. QuantaBlu Fluorogenic Peroxidase Substrates allow rapid kinetic ELISA assays to be performed with greater sensitivity than TMB and OPD substrates

Features of the QuantaBlu NS/K Fluorogenic Substrate Kit:

• More sensitive than TMB, OPD or ABTS substrates
• Large dynamic range (4 log peroxidase concentration range)
• Excellent stability: working solution is stable for 24 hours
• Large stokes shift; excitation/emission maxima of 325/420; range of 315-345/370-460
• Does not photobleach

A variety of substrates are available for detecting peroxidase activity in ELISA-based assays. Colorimetric substrates (e.g., TMB, OPD and ABTS) have been widely used for years. Each of these substrates varies greatly with respect to its performance characteristics such as detection sensitivity, working range and attainable signal-to-noise ratios.

QuantaBlu Fluorogenic Peroxidase Substrate meets all the requirements of an ideal substrate for use in peroxidase detection. QuantaBlu Substrate generates a blue fluorescent product upon reaction with peroxidase. The fluorescent product does not photobleach. Fluorometric-based detection overcomes the limitations of colorimetric substrate detection, which does not allow for quantitation of greater than four optical density units. QuantaBlu Substrate exhibits a flat baseline in assays, which facilitates low-level detection sensitivity and allows for high signal-to-noise ratios.

Related Products
QuantaBlu™ Fluorogenic Peroxidase Substrate Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionQuantaBlu NS/K Substrate (for non-stopped and kinetic assay)
Detection MethodFluorescence
Product LineQuantaBlu™
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase)
Product TypeELISA Substrate
Quantity280 mL
Unit SizeEach
Contents & Storage
Sufficient For: 275 mL of 1X working solution (2750 wells at 100 μL per well)
• QuantaBlu Substrate Solution, 250 mL
• QuantaBlu Peroxide Solution, 30 mL

Store at 4°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link